MagMAX™ Saliva gDNA Isolation Kit
MagMAX™ Saliva gDNA Isolation Kit
Applied Biosystems™

MagMAX™ Saliva gDNA Isolation Kit

The MagMAX Saliva gDNA Isolation Kit is a scalable and automatable magnetic bead-based sample preparation technology for the isolation ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A39059100
A39060500
Catalog number A39059
Price (USD)
668.00
Each
-
Add to cart
No. of Reactions:
100
Recurring order eligible. Learn more »
Price (USD)
668.00
Each
Add to cart
Ask our AI about this Product

The MagMAX Saliva gDNA Isolation Kit is a scalable and automatable magnetic bead-based sample preparation technology for the isolation of genomic DNA (gDNA) from fresh and stabilized saliva. The resulting purified gDNA is ideal for a variety of downstream molecular biology applications such as qPCR, next-generation sequencing, and microarray analysis. All components are also available for purchase separately.

Key features of the MagMAX Saliva gDNA Isolation Kit:
• Process up to 96 samples in 25 minutes with 5 minutes hands-on time
• Isolate gDNA from 200 μL–2 mL of saliva using simple protocols in SBS microtiter plates
• Skip sample and buffer volume normalization steps for hassle-free processing
• Download validated BindIt software automation scripts for KingFisher Duo Prime and KingFisher Flex purification systems

Isolate DNA from stabilized and freshly collected saliva
The MagMAX Saliva gDNA Isolation Kit isolates gDNA from 200 μL–2 mL of freshly collected saliva or stabilized saliva collected using a commercially available saliva collection device (see figure below). MagMAX Saliva gDNA isolation chemistry is compatible with the Oragene™ saliva collection devices from DNA Genotek, the SalivaGard™ saliva collection device from Biomatrica, and the GeneFiX™ Saliva DNA Collection Device from IsoHelix.

The MagMAX Saliva gDNA Isolation Kit maximizes gDNA yields by treating samples with an innovative lysis/binding solution to simultaneously lyse cells, denature proteins, and capture gDNA on high-capacity magnetic binding beads. The Saliva gDNA Wash I Solution removes proteins, detergents, and other impurities from the bound nucleic acid. The gDNA is then released from the beads using a low-TE elution solution.

Fast and hassle-free magnetic bead-based workflows
The MagMAX Saliva gDNA Isolation Kit combines multiple laborious sample processing steps together into a single step to minimize the time and effort typically required by other methods, without compromising yield and purity. Process a broad range of sample volume inputs simultaneously without sample normalization or reagent volume adjustments for hassle-free sample prep. Simply match your saliva input volumes to one of three standard input workflows (50 μL to 500 μL, up to 1 mL, up to 2 mL) and follow the instructions for semi-automated sample preparation using the KingFisher Duo Prime or KingFisher Flex purification systems.

Optimized for KingFisher purification systems
The MagMAX Saliva gDNA Isolation Kit is optimized for use on the moderate-throughput KingFisher Duo Prime (Cat. No. 5400110) and the higher-throughput KingFisher Flex (Cat. No. 5400630) purification systems. Users can process up to 500 μL of saliva in high-throughput capacity SBS 96-well deep-well microtiter plates using the KingFisher Flex system. For smaller sample batches, users can process 12 samples using the Duo Prime system. Larger sample input volumes (>500 μL, and 1–2 mL), which are more challenging to process, are purified in the larger sample volume capacity 24-well deep-well SBS plates for 24 samples on the Flex or 6 samples on the Duo Prime system. KingFisher instrument BindIt software scripts for all sample types and both Flex and Duo Prime systems can be downloaded below and installed onto the KingFisher instruments.

Related products
MagMAX Saliva gDNA Binding Beads
MagMAX Saliva gDNA Wash I Solution
MagMAX Saliva gDNA Lysis/Binding Solution
MagMAX Saliva gDNA Elution Solution

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
No. of Reactions100
Product LineMagMAX™
Product TypeSaliva gDNA Isolation Kit
Quantity100 Reactions
Shipping ConditionRoom Temperature
TargetGenomic DNA
Test Time25 min.
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
• 4.5 mL Binding Beads
• 110 mL Wash I Solution
• 55 mL Lysis/Binding Solution
• 12 mL Elution Solution

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2506030Certificate of AnalysisJun 27, 2025A39059
2503017Certificate of AnalysisApr 17, 2025A39060
2302020Certificate of AnalysisJan 27, 2025A39059
2402027Certificate of AnalysisJan 27, 2025A39059
2407028Certificate of AnalysisJan 26, 2025A39059
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

At lower temperatures, the components of the Lysis/Binding Solution and Wash I Solution may precipitate. To get rid of the precipitate, just warm the solutions to room temperature and invert bottles gently to solubilize the reagents before use.

Yes you can, but the gDNA yields will be very low.

No, this kit is specialized for freshly collected saliva and stabilized saliva samples only.

Yes, we offer the following kit components as stand-alone items to accommodate custom workflows or reagent volumes that have been optimized for your specific automation needs:
MagMAX Saliva gDNA Binding Beads: Cat. No. A39061
MagMAX Saliva gDNA Wash I Solution: Cat. No. A39062
MagMAX Saliva gDNA Lysis/Binding Solution: Cat. No. A39063
MagMAX Saliva gDNA Elution Solution: Cat. No. A39064

Unfortunately, we cannot give out the exact formulation, but it is similar to low TE (10 mM Tris base, 0.1 mM EDTA) and is compatible with microarray, PCR, and NGS downstream applications.

Share catalog number, name or link

1x1 image pixel for data collection