1x1 image pixel for data collection
TopVision Agarose
TopVision Agarose
Thermo Scientific™

TopVision Agarose

Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highlyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R0491100 g
R0492500 g
Catalog number R0491
Price (USD)
145.65
Online Exclusive
186.00
Save 40.35 (22%)
Each
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
186.00
Save 40.35 (22%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.

Highlights

• Optimal concentration between 0.4 to 5% in all typical buffer systems
• GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions, etc.)
• Low DNA/RNA binding
• Excellent gel transparency
• DNase and RNase free
• Suitable for RNA analysis

Applications

• Analytical electrophoresis of nucleic acids
• Preparative electrophoresis
• Blotting assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Melting Point88 °C
Product LineTopVision
Product TypeAgarose Gel
Quantity100 g
Validated ApplicationGel Electrophoresis
Separation Range100 to >30000 bp
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3269078Certificate of AnalysisJun 26, 2025R0491
3263866Certificate of AnalysisJun 18, 2025R0492
3258799Certificate of AnalysisJun 11, 2025R0491
3228723Certificate of AnalysisApr 30, 2025R0491
3202122Certificate of AnalysisApr 02, 2025R0491
5 results displayed, search above for a specific certificate

Safety Data Sheets

Los productos recomendados



Share catalog number, name or link