1x1 image pixel for data collection
Pro-Q™ Emerald 300 Glycoprotein Gel Stain Kit, with SYPRO™ Ruby Protein Gel Stain
Product Image
Invitrogen™

Pro-Q™ Emerald 300 Glycoprotein Gel Stain Kit, with SYPRO™ Ruby Protein Gel Stain

Molecular Probes' proprietary Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available forRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
P21855Promo Image1 kit
Catalog number P21855
Price (USD)
622.00
Each
In stock
Add to cart
Quantity:
1 kit
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
622.00
Each
Add to cart
Ask our AI about this Product
Molecular Probes' proprietary Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation, making these stains at least 50-fold more sensitive than standard fuchsin staining.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Blot Detection, In-Gel Detection
Detection MethodFluorescence
Product LinePRO-Q™, SYPRO™
Product TypeGlycoprotein Staining Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Target MoleculeProteins (Glycoproteins)
Label or DyeSYPRO Ruby, Pro-Q Emerald 300
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Frequently asked questions (FAQs)

Pro-Q Emerald Glycoprotein Gel Stain may show high background staining in Invitrogen NuPAGE Bis-Tris and Tris-Acetate gels, especially in combination with MES running buffer or in gels that are nearing their expiration date. The gel background increases with acrylamide density and gradient gels will show a gradual increase in background from the top to the bottom of the gel corresponding to the acrylamide gradient. Increasing the number of washes or modifying incubation times will not help to reduce this background. Better results will be obtained with Tris-Glycine or Tricine gels. If you wish to continue using Pro-Q Emerald stain with Invitrogen NuPAGE Bis-Tris gels, we recommend using recently purchased gels and MOPS running buffer. Glycoproteins will still be detected in gels with high background, but with reduced sensitivity.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Speckling of Pro-Q Emerald dye, especially with Pro-Q Emerald 300 stain, can occur as the Pro-Q Emerald dye ages, due to self-aggregation of the dye over time. Speckles can also form due to dye binding to contaminants from the staining container, solutions, or particles from the air or gloves. Non-dye speckles can also show up in the image from auto-fluorescent particles of dust, hair, glove powder, or clothing lint that falls on the gel or surface of the glass imaging plate. The better the imager is at focusing on surface features of the gel, the more speckles that are going to be visible.

To minimize the formation of speckles and other background debris, follow clean laboratory practices, use ultrapure water of >18 megohm-cm resistance to prepare solutions, rinse gloves in water to remove powder residue before touching gels, use lint-free wipes and wear a lab coat or avoid wearing clothing that generates a lot of lint, always rinse the staining container with ethanol and wipe out any residual dye before staining another gel, and always rinse and wipe down the glass imaging surface with ethanol and water before placing your gel down. Once speckles have been deposited on the gel, it is not possible to wash them off. When analyzing amounts of glycoprotein near the limit of detection, we advise running samples in the middle lanes of the gel.

Speckles will show up as sharp, tall spikes on 3D renditions of gel images. These spikes look distinct from 3D renditions of protein spots or bands. Some image analysis software packages have de-speckling algorithms that can easily identify and remove this type of pixelated noise.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Many total protein stains including SYPRO Ruby Gel Stain and Coomassie Blue stain will quench the Pro-Q Emerald signal. If you are staining your gels or blots with Pro-Q Emerald stain in containers that have previously been used for a total protein stain, you may be contaminating your gel with residue left on the staining dish from the total protein stain. Either use new containers, such as plastic weigh boats, designated containers for each stain, or rinse the container well in ethanol and wipe out any residual residue with a Kimwipe tissue.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

- Poor staining can be caused by the presence of primary amines, such as from Tris or glycine, that will also react with the aldehyde/ketone groups generated by periodate oxidation. This effectively caps the reactive groups, leaving no reactive sites for Pro-Q Emerald dye to bind. To remove any amine contamination, increase the volume or number of incubations in fresh fixative and then increase the volume or number of washes in wash buffer.
- Poor staining can also be due to inadequate removal of the periodic acid oxidation solution. Increase the volume or number of washes after the oxidation step to ensure adequate removal of periodic acid.
- Poor staining can also be due to inadequate oxidation of glycoprotein sugars. Increase the volume of periodic acid oxidation solution.

Note: Do not increase the number or incubation time of the periodic acid oxidation in excess of 30 minutes for small-format gels. Excessive periodic acid oxidation could result in increased staining of non-glycosylated proteins. In general, large format, unusually thick or very high percent acrylamide gels may require additional incubations or wash steps for optimal signal and sensitivity of staining with Pro-Q Emerald Glycoprotein Gel Stain.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

If the CandyCane standard and test glycoproteins are staining correctly, then the kit is performing well. Some very highly abundant proteins, such as albumin in serum and plasma, may stain lightly. The Pro-Q Emerald dye is covalently attached to sugar residues, so more post-staining washes will help to remove any non-covalently bound dye. Non-specific staining due to high abundance can be determined by post-staining with a total protein stain, such as SYPRO Ruby Protein Gel Stain. That being said, some proteins are actually heavily oxidized in the native state, and this carbonylation will be picked up by the Pro-Q Emerald reagent. Carbonylation of amino acids can be distinguished from glycosylation by performing the Pro-Q Emerald staining without the oxidation step. Under these conditions, the CandyCane marker bands will not be stained, but the carbonylated proteins will.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (9)

Citations & References
Abstract
Altered gingipain maturation in vimA- and vimE-defective isogenic mutants of Porphyromonas gingivalis.
Authors:Vanterpool E, Roy F, Sandberg L, Fletcher HM
Journal:Infect Immun
PubMed ID:15731033
'We have previously shown that gingipain activity in Porphyromonas gingivalis is modulated by the unique vimA and vimE genes. To determine if these genes had a similar phenotypic effect on protease maturation and activation, isogenic mutants defective in those genes were further characterized. Western blot analyses with antigingipain antibodies showed ... More
Ehrlichia chaffeensis expresses macrophage- and tick cell-specific 28-kilodalton outer membrane proteins.
Authors:Singu V, Liu H, Cheng C, Ganta RR
Journal:Infect Immun
PubMed ID:15618143
'Ehrlichia chaffeensis, a tick-transmitted rickettsial agent, causes human monocyte/macrophage-tropic ehrlichiosis. In this study, proteomic approaches were used to demonstrate host cell-specific antigenic expression by E. chaffeensis. The differentially expressed antigens include those from the 28-kDa outer membrane protein (p28-Omp) multigene locus. The proteins expressed in infected macrophages are the products ... More
Protein stains for proteomic applications: which, when, why?
Authors:Miller I, Crawford J, Gianazza E,
Journal:Proteomics
PubMed ID:16991193
'This review recollects literature data on sensitivity and dynamic range for the most commonly used colorimetric and fluorescent dyes for general protein staining, and summarizes procedures for the most common PTM-specific detection methods. It also compiles some important points to be considered in imaging and evaluation. In addition to theoretical ... More
Detection of glycoproteins in polyacrylamide gels and on electroblots using Pro-Q Emerald 488 dye, a fluorescent periodate Schiff-base stain.
Authors:Hart C, Schulenberg B, Steinberg TH, Leung WY, Patton WF
Journal:Electrophoresis
PubMed ID:12601726
Pro-Q Emerald 488 glycoprotein stain reacts with periodic acid-oxidized carbohydrate groups, generating a bright green-fluorescent signal on glycoproteins. The stain permits detection of less than 5-18 ng of glycoprotein per band, depending upon the nature and the degree of protein glycosylation, making it roughly 8-16-fold more sensitive than the standard ... More
Combining microscale solution-phase isoelectric focusing with Multiplexed Proteomics dye staining to analyze protein post-translational modifications.
Authors:Schulenberg B, Patton WF
Journal:Electrophoresis
PubMed ID:15300774
Previously, a strategy for rapidly identifying mitochondrial phosphoproteins was presented that involves prefractionating multisubunit complexes by sucrose gradient centrifugation, followed by sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis and selective staining of phosphoproteins and total protein with fluorescent dyes [1]. Though suitable for evaluating the mitochondrial proteome, which consists of numerous ... More
9 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection