Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R72501 | 1 mL |
Invitrogen 2.5 mM dNTP (2'-deoxynucleoside 5'-triphosphate) Mix consists of a solution of all four nucleotides, dATP, dCTP, dGTP, and dTTP, each at a concentration of 2.5 mM. It is supplied in purified water and neutralized to pH 8.0 with NaOH. 2.5 mM dNTP Mix is suitable for use in a wide variety of molecular applications.
Applications
• Traditional PCR
• Sequencing
• cDNA synthesis
• Nick translation
• TdT-tailing reactions
Share catalog number, name or link