ImmunoCAP™ Phadiatop™ 檢測在測量常見吸入性過敏原均勻混合物的 IgE 抗體,能幫助納入和排除異位性過敏的可能性,異位性過敏為一種對環境中蛋白質產生 IgE 誘發型反應之趨勢。ImmunoCAP Phadiatop Infant 針對小於 5 歲之孩童進行了優化,包括對該年齡族群來說相當重要的食物過敏原。1-5
這些檢測可幫助臨床醫師:
ImmunoCAP Phadiatop 檢測可在極少的限制下執行。不同於傳統的皮膚點刺測試 (SPT),ImmunoCAP 血液檢測:
幫助辨識出誘發症狀的過敏原和定量數值,建議以適當的 ImmunoCAP Specific IgE 全過敏原重新檢測樣本。
1.Directions for use.ImmunoCAP Phadiatop.
2.Eriksson, N.E., Allergy screening with Phadiatop and CAP Phadiatop in combination with a questionnaire in adults with asthma and rhinitis.Allergy, 1990.45(4): p. 285-92.
3.Fiocchi, A., et al., Differential diagnosis of IgE-mediated allergy in young children with wheezing or eczema symptoms using a single blood test.Ann Allergy Asthma Immunol, 2004.93(4): p. 328-33.
4.Nilsson C, et al.Pediatr Allergy Immunol.2012 Mar;23(2):159–65.
5.Ballardini, N., et al., ImmunoCAP Phadiatop Infant--a new blood test for detecting IgE sensitisation in children at 2 years of age.Allergy, 2006.61(3): p. 337-43.
6.Williams, P.B., C. Siegel, and J. Portnoy, Efficacy of a single diagnostic test for sensitization to common inhalant allergens.Ann Allergy Asthma Immunol, 2001.86(2): p. 196-202.
7.Halvorsen, R., et al., Phadiatop infant in the diagnosis of atopy in children with allergy-like symptoms.Int J Pediatr, 2009.2009: p. 460737.
8.Paganelli, R., et al., Specific IgE antibodies in the diagnosis of atopic disease.Clinical evaluation of a new in vitro test system, UniCAP, in six European allergy clinics.Allergy, 1998.53(8): p. 763-8.
9.Siles, R.I. and F.H.Hsieh, Allergy blood testing: A practical guide for clinicians.Cleve Clin J Med, 2011.78(9): p. 585-92.
10.Bonnelykke, K., C.B.Pipper, and H. Bisgaard, Sensitization does not develop in utero.J Allergy Clin Immunol, 2008.121(3): p. 646-51.
11.Belhocine, W., et al., Total serum tryptase levels are higher in young infants.Pediatr Allergy Immunol, 2011.22(6): p. 600-7.
12.Bacharier, L.B., et al., Diagnosis and treatment of asthma in childhood: a PRACTALL consensus report.Allergy, 2008.63(1): p. 5-34.
13.Walsh, J. and N. O'Flynn, Diagnosis and assessment of food allergy in children and young people in primary care and community settings: NICE clinical guideline.The British journal of general practice : the journal of the Royal College of General Practitioners, 2011.61(588): p. 473-475.
14.ImmunoDiagnostics Statment, No Biotin Interference in ImmunoCAPTM Assays.Uppsala, Jan 31 2018.