Maximize your RT-qPCR success with optimized starting material
RT-qPCR requires both high-quality RNA as starting material and optimal conversion of the RNA to a DNA copy for accurate real-time PCR detection (qPCR). When an RNA quantitation step is added you’ll be better able to optimize your qPCR analysis.
Learn more about optimizing each step in the workflow by clicking on the tabs below.
The sensitivity and accuracy of RT-qPCR detection depends on the quality and quantity of your input cDNA, which in turn depends on the quality and integrity of the isolated RNA. One important step for avoiding false positives and background is the removal of genomic DNA.
Invitrogen Ambion RNA purification products from Thermo Fisher Scientific help simplify RNA Extraction for Real-Time PCR. You can also capture your RT-qPCR results directly from cells with Cells-to-CT.
Compare RNA isolation kits for fresh and frozen cells:
 Order NowOrder NowOrder NowOrder Now
 Rapid & reliable methodHigh-throughput purificationmicroRNA from most samplesRNA from precious micro-sized samples
 PureLink RNA Mini KitMagMAX-96 Total RNA Isolation KitmirVana miRNA Isolation KitRNAqueous-Micro Kit
RNA type(s) isolatedLarge RNA molecules only (mRNA and rRNA)Large RNA molecules only (mRNA and rRNA)Small & large RNA molecules (microRNA, tRNA, mRNA, rRNA)Small & large RNA molecules (microRNA, tRNA, mRNA, rRNA)
Isolation methodFast, convenient silica columnScalable, flexible format with magnetic beadsHighest purity and convenience; includes both organic extraction and silica columnFast, convenient low elution silica column
Preparation time20 minutes<45 minutes30 minutes15 minutes
Amount of starting material10 mg to 100 mg of tissue
Up to 5 x 107 cells
Up to 10 mg of tissue
Up to 100,000 cells
Up to 100 mg of tissue
Up to 1 x 107 cells
Up to 10 mg of tissue
Up to 100,000 cells
Kit size (preps)50964050
The Qubit™ and Quant-iT™ RNA assays provide specific, sensitive RNA and microRNA quantification, even at low concentrations, with minimal interference from contaminants.
Unlike UV absorbance readings, which are less discriminate, these assays distinguish between DNA, RNA, proteins, and free nucleotides with higher sensitivity than UV absorbance readings, making it possible to measure low-abundance samples. In addition, an intuitive format makes it easier and faster to get your quantifation results.
Find out more about Qubit™ and Quant-iT™ technology for your precious samples.
1–20 samples: Qubit™ Assays for use on Qubit™ Fluorometer
Catalog # Name Size Price (USD) Qty
Q10210 Qubit™ RNA BR Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q10211 Qubit™ RNA BR Assay Kit, 500 assays Each
408.65
Online Exclusive
431.00
Save 22.35 (5%)
Q32852 Qubit™ RNA HS Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q32855 Qubit™ RNA HS Assay Kit, 500 assays Each
406.65
Online Exclusive
430.00
Save 23.35 (5%)
Q32880 Qubit™ microRNA Assay Kit, 100 assays Each
139.65
Online Exclusive
146.00
Save 6.35 (4%)
Q32881 Qubit™ microRNA Assay Kit, 500 assays Each
421.65
Online Exclusive
444.00
Save 22.35 (5%)
Q33216 Qubit™ 3 Fluorometer Each -
20–2,000 samples: Quant-iT™ Assays for use on fluorescent microplate reader
Catalog # Name Size Price (USD) Qty
Q33140 Quant-iT™ RNA Assay Kit, 1 kit Each
580.65
Online Exclusive
624.00
Save 43.35 (7%)
Q10213 Quant-iT™ RNA Assay Kit, broad range, 1,000 assays Each
592.65
Online Exclusive
624.00
Save 31.35 (5%)
Q32882 Quant-iT™ microRNA Assay Kit Each
735.65
Online Exclusive
758.00
Save 22.35 (3%)
For reliable generation of first strand cDNA, SuperScript IV VILO Master Mix brings you proven SuperScript IV Reverse Transcriptase formulated in an enhanced buffer system.  Reduce amplification bias from low- or high-abundance gene products, and capture data that exhibits superior linearity across a broad range of input material. 
The SuperScript IV VILO Master Mix is available in a convenient single-tube format, facilitating a simplified, time-saving workflow and helping to reduce the risk of contamination.
Find out more about the SuperScript IV VILO Master Mix ›
cDNA Synthesis Master Mixes for Real Time RT-PCR
Catalog # Name Size Price (USD) Qty
11756050 SuperScript™ IV VILO™ Master Mix, Each Each
776.65
Online Exclusive
828.00
Save 51.35 (6%)
11756500 SuperScript™ IV VILO™ Master Mix, Each Each
5,684.65
Online Exclusive
6,060.00
Save 375.35 (6%)
11766050 SuperScript™ IV VILO™ Master Mix with ezDNase™ Enzyme, Each Each
817.65
Online Exclusive
872.00
Save 54.35 (6%)
11766500 SuperScript™ IV VILO™ Master Mix with ezDNase™ Enzyme, Each Each
6,125.65
Online Exclusive
6,530.00
Save 404.35 (6%)
11766051 ezDNase™ Enzyme Each
288.65
Online Exclusive
311.00
Save 22.35 (7%)
Reverse Transcriptase
Catalog # Name Size Price (USD) Qty
18090010 SuperScript™ IV Reverse Transcriptase, 2,000 Units Each
133.65
Online Exclusive
137.00
Save 3.35 (2%)
18090050 SuperScript™ IV Reverse Transcriptase, 10,000 Units Each
497.65
Online Exclusive
530.00
Save 32.35 (6%)
18090200 SuperScript™ IV Reverse Transcriptase, 4 x 10,000 Units Each
1,763.65
Online Exclusive
1,876.00
Save 112.35 (6%)
The sensitivity and accuracy of RT-qPCR detection depends on the quality and quantity of your input cDNA, which in turn depends on the quality and integrity of the isolated RNA. One important step for avoiding false positives and background is the removal of genomic DNA.
Invitrogen Ambion RNA purification products from Thermo Fisher Scientific help simplify RNA Extraction for Real-Time PCR. You can also capture your RT-qPCR results directly from cells with Cells-to-CT.
Compare RNA isolation kits for fresh and frozen cells:
 Order NowOrder NowOrder NowOrder Now
 Rapid & reliable methodHigh-throughput purificationmicroRNA from most samplesRNA from precious micro-sized samples
 PureLink RNA Mini KitMagMAX-96 Total RNA Isolation KitmirVana miRNA Isolation KitRNAqueous-Micro Kit
RNA type(s) isolatedLarge RNA molecules only (mRNA and rRNA)Large RNA molecules only (mRNA and rRNA)Small & large RNA molecules (microRNA, tRNA, mRNA, rRNA)Small & large RNA molecules (microRNA, tRNA, mRNA, rRNA)
Isolation methodFast, convenient silica columnScalable, flexible format with magnetic beadsHighest purity and convenience; includes both organic extraction and silica columnFast, convenient low elution silica column
Preparation time20 minutes<45 minutes30 minutes15 minutes
Amount of starting material10 mg to 100 mg of tissue
Up to 5 x 107 cells
Up to 10 mg of tissue
Up to 100,000 cells
Up to 100 mg of tissue
Up to 1 x 107 cells
Up to 10 mg of tissue
Up to 100,000 cells
Kit size (preps)50964050
The Qubit™ and Quant-iT™ RNA assays provide specific, sensitive RNA and microRNA quantification, even at low concentrations, with minimal interference from contaminants.
Unlike UV absorbance readings, which are less discriminate, these assays distinguish between DNA, RNA, proteins, and free nucleotides with higher sensitivity than UV absorbance readings, making it possible to measure low-abundance samples. In addition, an intuitive format makes it easier and faster to get your quantifation results.
Find out more about Qubit™ and Quant-iT™ technology for your precious samples.
1–20 samples: Qubit™ Assays for use on Qubit™ Fluorometer
Catalog # Name Size Price (USD) Qty
Q10210 Qubit™ RNA BR Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q10211 Qubit™ RNA BR Assay Kit, 500 assays Each
408.65
Online Exclusive
431.00
Save 22.35 (5%)
Q32852 Qubit™ RNA HS Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q32855 Qubit™ RNA HS Assay Kit, 500 assays Each
406.65
Online Exclusive
430.00
Save 23.35 (5%)
Q32880 Qubit™ microRNA Assay Kit, 100 assays Each
139.65
Online Exclusive
146.00
Save 6.35 (4%)
Q32881 Qubit™ microRNA Assay Kit, 500 assays Each
421.65
Online Exclusive
444.00
Save 22.35 (5%)
Q33216 Qubit™ 3 Fluorometer Each -
20–2,000 samples: Quant-iT™ Assays for use on fluorescent microplate reader
Catalog # Name Size Price (USD) Qty
Q33140 Quant-iT™ RNA Assay Kit, 1 kit Each
580.65
Online Exclusive
624.00
Save 43.35 (7%)
Q10213 Quant-iT™ RNA Assay Kit, broad range, 1,000 assays Each
592.65
Online Exclusive
624.00
Save 31.35 (5%)
Q32882 Quant-iT™ microRNA Assay Kit Each
735.65
Online Exclusive
758.00
Save 22.35 (3%)
For reliable generation of first strand cDNA, SuperScript IV VILO Master Mix brings you proven SuperScript IV Reverse Transcriptase formulated in an enhanced buffer system.  Reduce amplification bias from low- or high-abundance gene products, and capture data that exhibits superior linearity across a broad range of input material. 
The SuperScript IV VILO Master Mix is available in a convenient single-tube format, facilitating a simplified, time-saving workflow and helping to reduce the risk of contamination.
Find out more about the SuperScript IV VILO Master Mix ›
cDNA Synthesis Master Mixes for Real Time RT-PCR
Catalog # Name Size Price (USD) Qty
11756050 SuperScript™ IV VILO™ Master Mix, Each Each
776.65
Online Exclusive
828.00
Save 51.35 (6%)
11756500 SuperScript™ IV VILO™ Master Mix, Each Each
5,684.65
Online Exclusive
6,060.00
Save 375.35 (6%)
11766050 SuperScript™ IV VILO™ Master Mix with ezDNase™ Enzyme, Each Each
817.65
Online Exclusive
872.00
Save 54.35 (6%)
11766500 SuperScript™ IV VILO™ Master Mix with ezDNase™ Enzyme, Each Each
6,125.65
Online Exclusive
6,530.00
Save 404.35 (6%)
11766051 ezDNase™ Enzyme Each
288.65
Online Exclusive
311.00
Save 22.35 (7%)
Reverse Transcriptase
Catalog # Name Size Price (USD) Qty
18090010 SuperScript™ IV Reverse Transcriptase, 2,000 Units Each
133.65
Online Exclusive
137.00
Save 3.35 (2%)
18090050 SuperScript™ IV Reverse Transcriptase, 10,000 Units Each
497.65
Online Exclusive
530.00
Save 32.35 (6%)
18090200 SuperScript™ IV Reverse Transcriptase, 4 x 10,000 Units Each
1,763.65
Online Exclusive
1,876.00
Save 112.35 (6%)
Citations
Deben, C et al. (2013) Expression analysis on archival material revisited: isolation and quantification of RNA extracted from FFPE samples.Diagn Mol Pathol 22(1):59-64.
“A fluorometric analysis is more suitable for quantification of RNA samples extracted from FFPE tissue  compared with spectrophotometric analysis." (Qubit® RNA quantitation)
Simbolo, M et al. (2013) DNA qualification workflow for next generation sequencing of histopathological samples.PLoS One 8(6):e62692.
"NanoDrop measured DNA concentrations higher than Qubit and its consistency with dsDNA quantification by qPCR was limited to high molecular weight DNA from FF samples and cell lines, where total DNA and dsDNA quantity virtually coincide. In partially degraded DNA from FFPE samples, only Qubit proved highly reproducible and consistent with qPCR measurements.”
For Research Use Only. Not for use in diagnostic procedures.