bf5a- Apps/Techniques/Products/Resources/Contact Us
Modern cutting-edge metals are increasingly engineered at the nanoscale to enhance their durability, reliability, and cost. Even traditional processes are now augmented with microscopic inspection to determine the resulting material’s elemental and structural composition.
In particular, the effective production of metals requires precise control of inclusions and precipitates. Depending on their consistency and distribution, these can either strengthen the material or act as contaminants, greatly impacting quality and lifetime. These microscopic properties can include;
  • Nano-precipitates formed during rolling, annealing or hot pressing
  • Nanoscale morphological changes (dislocations, crack initiation sites)
  • Grain boundaries
  • Oxide inclusions that cause casting interruptions in steelmaking
Historically, researchers have used optical microscopy to rate the size and number of inclusions, but this method does not provide any elemental information. Even optical emission spectroscopy, which can determine the elemental ratios of inclusions, does not accurately characterize the shape and composition of individual inclusions. Electron microscopy techniques have also been used for metal analysis, with scanning electron microscopy (SEM) capable of visualizing larger oxide inclusions, whereas transmission electron microscopy (TEM) is generally required to study features smaller than 100 nm. TEM analysis, however, has previously required manual particle counting and analysis, limiting the amount of data that could be collected to several dozen particles per day.
Stainless steel medical device sample prepared by PFIB milling.
Stainless steel medical device sample generated with PFIB milling, with total dimensions of 55 x 70 μm. The red box indicates the amount of area that could be prepared in the same amount of time with a typical gallium FIB.
Thermo Fisher Scientific provides a range of electron microscopy solutions that make metal analysis not only more informative but also far more rapid. Thanks to our unique automation capabilities, a thorough overview of the elemental and structural composition of hundreds, if not thousands, of precipitates is possible in a manner of hours, as compared to the few dozen that would be found in a day of manual analysis. Not only is statistical information on the bulk available, but individual precipitates can also be seen with high detail, providing a multi-scale overview of the metal.
Our robust, automated instruments can perform a variety of critical tasks including:
Zirconium alloy sample, analyzed with electron backscatter diffraction to produce a 3D microstructural reconstruction.
3D microstructural information provided by electron backscatter diffraction (EBSD) of a zirconium alloy sample reconstructed from 400 slices. Sample courtesy of the University of Manchester.
Resources
SEM images
Low-carbon, aluminum-killed steel sample observed by Phenom ParticleX Steel Desktop SEM
Low-carbon, aluminum-killed steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows a cluster of several micron-sized alumina inclusions.
Titanium-stabilized, ultra-low-carbon steel sample observed by Phenom ParticleX Steel Desktop SEM
Titanium-stabilized, ultra-low-carbon steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows cubic titanium nitride precipitating on top of an oxide inclusion.
Calcium-treated steel sample observed by Phenom ParticleX Steel Desktop SEM
Calcium-treated steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows MgO.Al2O3 spinel phase (dark) forming inside a calcium aluminate inclusion.
Calcium-treated steel ternary diagram
Calcium-treated steel was scanned by a Phenom ParticleX Steel Desktop SEM over 60 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the composition distribution of calcium aluminates and calcium sulfides in this steel alloy.
Calcium-treated steel scanned
Calcium-treated steel was scanned by a Phenom ParticleX Steel Desktop SEM over 60 mm2 to characterize non-metallic inclusions. This particle classification table shows the count and average composition of over 1,500 inclusions that were identified.
Calcium-treated steel sample
Calcium-treated steel sample observed by a Phenom ParticleX Steel Desktop SEM. Backscattered imaging (left) and EDS mapping show a compound calcium sulfide and calcium aluminate inclusion. On the EDS maps, calcium is shown in yellow and aluminum in blue.
Forged steel cylinder polished section observed by Phenom ParticleX Steel. Backscattered imaging reveals the spatial distribution of large (> 5 μm) and small (< 1 μm) titanium rich particles. Sample courtesy of GKN Aerospace.
Forged steel cylinder polished section observed by Phenom ParticleX Steel. Backscattered imaging reveals the spatial distribution of large (> 5 μm) and small (< 1 μm) titanium rich particles. Sample courtesy of GKN Aerospace.
Forged steel cylinder polished section was scanned by ParticleX Steel over 50 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the size and composition distribution of titanium sulfides and titanium nitrides in this steel alloy. Sample courtesy of GKN Aerospace.
Forged steel cylinder polished section was scanned by ParticleX Steel over 50 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the size and composition distribution of titanium sulfides and titanium nitrides in this steel alloy. Sample courtesy of GKN Aerospace.
Forged steel cylinder surface observed by Axia ChemiSEM. This area has been machined which exposed large non-metallic phases. ChemiSEM EDS mapping confirmed that the particles are titanium rich. Sample courtesy of GKN Aerospace.
Forged steel cylinder surface observed by Axia ChemiSEM. This area has been machined which exposed large non-metallic phases. ChemiSEM EDS mapping confirmed that the particles are titanium rich. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm<sup>2</sup> to characterize particles brighter than the base metal. This Fe-Mn-Cu ternary diagram shows the chemical distribution of 65k bright phase particles. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal. This Fe-Mn-Cu ternary diagram shows the chemical distribution of 65k bright phase particles. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal.  This particle table shows the majority of the 65k particles contain a portion of iron, manganese and copper. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal. This particle table shows the majority of the 65k particles contain a portion of iron, manganese and copper. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel to characterize heavy metal particles. Backscattered electron image reveals bright phase particles which have a higher average atomic weight than the base metal. Sample courtesy of GKN Aerospace.
Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel to characterize heavy metal particles. Backscattered electron image reveals bright phase particles which have a higher average atomic weight than the base metal. Sample courtesy of GKN Aerospace.
XPS images
Forged steel cylinder surface analyzed by the Thermo Scientific Nexsa G2 Surface Analysis System. Optical micrograph reveals some of the machining detail on this heat treated sample. Sample courtesy of GKN Aerospace.
Forged steel cylinder surface analyzed by the Thermo Scientific Nexsa G2 Surface Analysis System. Optical micrograph reveals some of the machining detail on this heat treated sample. Sample courtesy of GKN Aerospace.
Forged steel cylinder surface analyzed by XPS with depth profiling.  It reveals the passivated layer of chromium oxide with the unoxidized steel beneath. Sample courtesy of GKN Aerospace.
Forged steel cylinder surface analyzed by XPS with depth profiling. It reveals the passivated layer of chromium oxide with the unoxidized steel beneath. Sample courtesy of GKN Aerospace.
Precipitates of niobium carbide in a high-strength, low-alloy steel were analyzed with a Talos F200X (S)TEM and Automated Particle Workflow (APW). The two regions represent different locations on the same coil, where the steel with finer precipitates (average 9 nm, left) yielded a higher strength than the steel with larger precipitates (average 12 nm, right).
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image, b) zinc EDS map, c) zinc particle segmentation. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image, b) zinc EDS map, c) zinc particle segmentation. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) magnesium particle segmentation, b) zinc particle segmentation, and c) Co-Located magnesium and zinc compounds shown in orange. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) magnesium particle segmentation, b) zinc particle segmentation, and c) Co-Located magnesium and zinc compounds shown in orange. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image and b) corresponding APW particle map of magnesium and zinc compounds. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image and b) corresponding APW particle map of magnesium and zinc compounds. Sample courtesy of University of Manchester and University of Trento.
Talos F200X S/TEM analysis of high strength low alloy steel carbon replica. HAADF STEM image shows precipitates and grain boundaries as a lighter color on a dark background. Sample courtesy of OCAS.
Talos F200X S/TEM analysis of high strength low alloy steel carbon replica. HAADF STEM image shows precipitates and grain boundaries as a lighter color on a dark background. Sample courtesy of OCAS.
Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Zoom area of the segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Zoom area of the segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Chart shows compound particle (TiN + NbC) size distribution, and segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Chart shows compound particle (TiN + NbC) size distribution, and segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
Talos F200X S/TEM analysis of additively manufactured 17-4 PH stainless steel showing: a) HAADF STEM image of a precipitate in the TEM lamella, and b) manual EDS mapping of the complex precipitate which contains a majority of MnSi oxide at the center and precipitates of Cu and NbN on the perimeter. Sample courtesy of University of Connecticut.
Talos F200X S/TEM analysis of additively manufactured 17-4 PH stainless steel showing: a) HAADF STEM image of a precipitate in the TEM lamella, and b) manual EDS mapping of the complex precipitate which contains a majority of MnSi oxide at the center and precipitates of Cu and NbN on the perimeter. Sample courtesy of University of Connecticut.
High resolution APW analysis showing EDS maps of Silicon (green) and Niobium (red) or Avizo particle quantification of silicon particles. Sample courtesy of University of Connecticut
High resolution APW analysis showing EDS maps of Silicon (green) and Niobium (red) or Avizo particle quantification of silicon particles. Sample courtesy of University of Connecticut
High resolution APW analysis showing compounds precipitates of silicon (yellow) and niobium (blue). Sample courtesy of University of Connecticut.
High resolution APW analysis showing compounds precipitates of silicon (yellow) and niobium (blue). Sample courtesy of University of Connecticut.
SEM videos
Video Player is loading.
Current Time 0:00
Duration 1:05
Loaded: 15.00%
Stream Type LIVE
Remaining Time 1:05
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Phenom ParticleX Steel Desktop SEM inclusion analysis short demonstration.
    Video Player is loading.
    Current Time 0:00
    Duration 6:23
    Loaded: 2.57%
    Stream Type LIVE
    Remaining Time 6:23
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      ParticleX Steel Desktop SEM - Workflow introduction.
      Video Player is loading.
      Current Time 0:00
      Duration 1:22
      Loaded: 11.95%
      Stream Type LIVE
      Remaining Time 1:22
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Axia ChemiSEM provides high-quality imaging of steel samples to aid in the production of high-value steels. 
        Video Player is loading.
        Current Time 0:00
        Duration 0:36
        Loaded: 27.78%
        Stream Type LIVE
        Remaining Time 0:36
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Axia ChemiSEM identifies composition on-the-fly
          TEM videos
          Aluminum 2099 alloy lamella characterization of Cu and Zr precipitates by APW
          Video Player is loading.
          Current Time 0:00
          Duration 1:10
          Loaded: 14.23%
          Stream Type LIVE
          Remaining Time 1:10
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            Nickel superalloy sample characterizingr titanium nitride nano particles by APW.
            Video Player is loading.
            Current Time 0:00
            Duration 1:04
            Loaded: 15.37%
            Stream Type LIVE
            Remaining Time 1:04
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              HSLA steel lamella characterization of Nb precipitates by Automated Particle Workflow (APW).
              Video Player is loading.
              Current Time 0:00
              Duration 0:19
              Loaded: 50.96%
              Stream Type LIVE
              Remaining Time 0:19
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • default, selected
                3D EDS TEM tomography of precipitates in an AlMgSi alloy.
                Video Player is loading.
                Current Time 0:00
                Duration 0:53
                Loaded: 18.43%
                Stream Type LIVE
                Remaining Time 0:53
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  High resolution APW showing complex features in additively manufactured stainless steel.
                  Video Player is loading.
                  Current Time 0:00
                  Duration 2:02
                  Loaded: 8.16%
                  Stream Type LIVE
                  Remaining Time 2:02
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • default, selected
                    Maps and Avizo2D recordings (left and right) running side by side during an acquisition.
                    Webinars
                    Video Player is loading.
                    Current Time 0:00
                    Duration 10:03
                    Loaded: 1.63%
                    Stream Type LIVE
                    Remaining Time 10:03
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      Webinar: Nanoparticle Characterization by Automated TEM.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 14:35
                      Loaded: 1.13%
                      Stream Type LIVE
                      Remaining Time 14:35
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • en (Main), selected
                        Webinar: Correlative Microscopy for Aerospace and Defense Industries
                        TEM Articles
                        Nanoscale origins of the oriented precipitation of Ti3Al in Ti\\Al systems
                        Hao Wu, Guohua Fan, Lin Geng, Xiping Cui, Meng Huang
                        DOI link
                        Effect of heat treatments on microstructural evolution of additively manufactured and wrought 17-4PH stainless steel
                        Yu Sun, Rainer J. Hebert, Mark Aindow
                        DOI link
                        Coherency strains of H-phase precipitates and their influence on functional properties of nickel-titanium-hafnium shape memory alloys
                        Behnam Amin-Ahmadi,⁎, Joseph G. Pauza, Ali Shamimi, Tom W. Duerig, Ronald D. Noebe, Aaron P. Stebner
                        DOI link
                        Effect of laser scan length on the microstructure of additively manufactured 17-4PH stainless steel thin-walled parts
                        Yu Sun, Rainer J. Hebert, Mark Aindow
                        DOI link
                        Non-metallic inclusions in 17-4PH stainless steel parts produced by selective laser melting
                        Yu Sun, Rainer J. Hebert, Mark Aindow
                        DOI link
                        FIB-SEM Articles
                        Joachim Mayer, RWTH Aachen
                        “Formation of White Etching Areas in SAE 52100 Bearing Steel under Rolling Contact Fatigue − Influence of Diffusible Hydrogen”
                        M. Oezel, A. Schwedt, T. Janitzky, R. Kelley, C.Bouchet-Marquis, L. Pullan, C. Broeckmann, J. Mayer
                        Wear, Volumes 414-415, November 2018, Pages 352-365.
                        DOI link
                        Philip Withers, University of Manchester
                        “Industrial Gear Oils: Tribological Performance and Subsurface Changes”
                        Aduragbemi Adebogun, Robert Hudson, Angela Breakspear, Chris Warrens, Ali Gholinia, Allan Matthews, Philip Withers Tribology Letters (2018) 66:65.
                        DOI link
                        Jun Tan, Shenyang National Laboratory for Materials Science
                        “Insight into atmospheric pitting corrosion of carbon steel via a dual-beam FIB/SEM system associated with high-resolution TEM”
                        Corrosion Science 152 (2019) 226–233.
                        DOI link
                        Yu-Lung Chiu, University of Birmingham
                        “Micro-tensile strength of a welded turbine disc superalloy”
                        K.M. Oluwasegun, C.Cooper, Y.L.Chiu, I.P.Jones, H.Y.Li, G.Baxter
                        Materials Science & Engineering A 596 (2014) 229–235.
                        DOI link
                        Chris Pistorius, Carnegie Mellon University
                        “Application of Plasma FIB to Analyze a Single Oxide Inclusion in Steel”
                        D. Kumar, N.T. Nuhfer, M.E. Ferreira and P.C. Pistorius
                        Metallurgical and Materials Transactions B, Volume 50B, June 2019, Pages 1124-1127.
                        DOI link
                        SEM images
                        Low-carbon, aluminum-killed steel sample observed by Phenom ParticleX Steel Desktop SEM
                        Low-carbon, aluminum-killed steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows a cluster of several micron-sized alumina inclusions.
                        Titanium-stabilized, ultra-low-carbon steel sample observed by Phenom ParticleX Steel Desktop SEM
                        Titanium-stabilized, ultra-low-carbon steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows cubic titanium nitride precipitating on top of an oxide inclusion.
                        Calcium-treated steel sample observed by Phenom ParticleX Steel Desktop SEM
                        Calcium-treated steel sample observed by Phenom ParticleX Steel Desktop SEM. Backscattered imaging shows MgO.Al2O3 spinel phase (dark) forming inside a calcium aluminate inclusion.
                        Calcium-treated steel ternary diagram
                        Calcium-treated steel was scanned by a Phenom ParticleX Steel Desktop SEM over 60 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the composition distribution of calcium aluminates and calcium sulfides in this steel alloy.
                        Calcium-treated steel scanned
                        Calcium-treated steel was scanned by a Phenom ParticleX Steel Desktop SEM over 60 mm2 to characterize non-metallic inclusions. This particle classification table shows the count and average composition of over 1,500 inclusions that were identified.
                        Calcium-treated steel sample
                        Calcium-treated steel sample observed by a Phenom ParticleX Steel Desktop SEM. Backscattered imaging (left) and EDS mapping show a compound calcium sulfide and calcium aluminate inclusion. On the EDS maps, calcium is shown in yellow and aluminum in blue.
                        Forged steel cylinder polished section observed by Phenom ParticleX Steel. Backscattered imaging reveals the spatial distribution of large (> 5 μm) and small (< 1 μm) titanium rich particles. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder polished section observed by Phenom ParticleX Steel. Backscattered imaging reveals the spatial distribution of large (> 5 μm) and small (< 1 μm) titanium rich particles. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder polished section was scanned by ParticleX Steel over 50 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the size and composition distribution of titanium sulfides and titanium nitrides in this steel alloy. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder polished section was scanned by ParticleX Steel over 50 mm2 to characterize non-metallic inclusions. This ternary diagram reveals the size and composition distribution of titanium sulfides and titanium nitrides in this steel alloy. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder surface observed by Axia ChemiSEM. This area has been machined which exposed large non-metallic phases. ChemiSEM EDS mapping confirmed that the particles are titanium rich. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder surface observed by Axia ChemiSEM. This area has been machined which exposed large non-metallic phases. ChemiSEM EDS mapping confirmed that the particles are titanium rich. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm<sup>2</sup> to characterize particles brighter than the base metal. This Fe-Mn-Cu ternary diagram shows the chemical distribution of 65k bright phase particles. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal. This Fe-Mn-Cu ternary diagram shows the chemical distribution of 65k bright phase particles. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal.  This particle table shows the majority of the 65k particles contain a portion of iron, manganese and copper. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel over 568 mm2 to characterize particles brighter than the base metal. This particle table shows the majority of the 65k particles contain a portion of iron, manganese and copper. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel to characterize heavy metal particles. Backscattered electron image reveals bright phase particles which have a higher average atomic weight than the base metal. Sample courtesy of GKN Aerospace.
                        Friction stir welded aerospace aluminum alloy was scanned by ParticleX Steel to characterize heavy metal particles. Backscattered electron image reveals bright phase particles which have a higher average atomic weight than the base metal. Sample courtesy of GKN Aerospace.
                        XPS images
                        Forged steel cylinder surface analyzed by the Thermo Scientific Nexsa G2 Surface Analysis System. Optical micrograph reveals some of the machining detail on this heat treated sample. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder surface analyzed by the Thermo Scientific Nexsa G2 Surface Analysis System. Optical micrograph reveals some of the machining detail on this heat treated sample. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder surface analyzed by XPS with depth profiling.  It reveals the passivated layer of chromium oxide with the unoxidized steel beneath. Sample courtesy of GKN Aerospace.
                        Forged steel cylinder surface analyzed by XPS with depth profiling. It reveals the passivated layer of chromium oxide with the unoxidized steel beneath. Sample courtesy of GKN Aerospace.
                        Precipitates of niobium carbide in a high-strength, low-alloy steel were analyzed with a Talos F200X (S)TEM and Automated Particle Workflow (APW). The two regions represent different locations on the same coil, where the steel with finer precipitates (average 9 nm, left) yielded a higher strength than the steel with larger precipitates (average 12 nm, right).
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image, b) zinc EDS map, c) zinc particle segmentation. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image, b) zinc EDS map, c) zinc particle segmentation. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) magnesium particle segmentation, b) zinc particle segmentation, and c) Co-Located magnesium and zinc compounds shown in orange. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) magnesium particle segmentation, b) zinc particle segmentation, and c) Co-Located magnesium and zinc compounds shown in orange. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image and b) corresponding APW particle map of magnesium and zinc compounds. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of Aluminum 7075 aerospace alloy showing a) HAADF STEM image and b) corresponding APW particle map of magnesium and zinc compounds. Sample courtesy of University of Manchester and University of Trento.
                        Talos F200X S/TEM analysis of high strength low alloy steel carbon replica. HAADF STEM image shows precipitates and grain boundaries as a lighter color on a dark background. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis of high strength low alloy steel carbon replica. HAADF STEM image shows precipitates and grain boundaries as a lighter color on a dark background. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Zoom area of the segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Zoom area of the segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Chart shows compound particle (TiN + NbC) size distribution, and segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis via Automated Particle Workflow of high strength low alloy steel carbon replica. Chart shows compound particle (TiN + NbC) size distribution, and segmented particle map shows titanium in gold, niobium in pink and where they overlap is orange. Sample courtesy of OCAS.
                        Talos F200X S/TEM analysis of additively manufactured 17-4 PH stainless steel showing: a) HAADF STEM image of a precipitate in the TEM lamella, and b) manual EDS mapping of the complex precipitate which contains a majority of MnSi oxide at the center and precipitates of Cu and NbN on the perimeter. Sample courtesy of University of Connecticut.
                        Talos F200X S/TEM analysis of additively manufactured 17-4 PH stainless steel showing: a) HAADF STEM image of a precipitate in the TEM lamella, and b) manual EDS mapping of the complex precipitate which contains a majority of MnSi oxide at the center and precipitates of Cu and NbN on the perimeter. Sample courtesy of University of Connecticut.
                        High resolution APW analysis showing EDS maps of Silicon (green) and Niobium (red) or Avizo particle quantification of silicon particles. Sample courtesy of University of Connecticut
                        High resolution APW analysis showing EDS maps of Silicon (green) and Niobium (red) or Avizo particle quantification of silicon particles. Sample courtesy of University of Connecticut
                        High resolution APW analysis showing compounds precipitates of silicon (yellow) and niobium (blue). Sample courtesy of University of Connecticut.
                        High resolution APW analysis showing compounds precipitates of silicon (yellow) and niobium (blue). Sample courtesy of University of Connecticut.
                        SEM videos
                        Video Player is loading.
                        Current Time 0:00
                        Duration 1:05
                        Loaded: 15.00%
                        Stream Type LIVE
                        Remaining Time 1:05
                        1x
                          • Chapters
                          • descriptions off, selected
                          • captions off, selected
                          • en (Main), selected
                          Phenom ParticleX Steel Desktop SEM inclusion analysis short demonstration.
                          Video Player is loading.
                          Current Time 0:00
                          Duration 6:23
                          Loaded: 2.57%
                          Stream Type LIVE
                          Remaining Time 6:23
                          1x
                            • Chapters
                            • descriptions off, selected
                            • captions off, selected
                            • en (Main), selected
                            ParticleX Steel Desktop SEM - Workflow introduction.
                            Video Player is loading.
                            Current Time 0:00
                            Duration 1:22
                            Loaded: 11.95%
                            Stream Type LIVE
                            Remaining Time 1:22
                            1x
                              • Chapters
                              • descriptions off, selected
                              • captions off, selected
                              • en (Main), selected
                              Axia ChemiSEM provides high-quality imaging of steel samples to aid in the production of high-value steels. 
                              Video Player is loading.
                              Current Time 0:00
                              Duration 0:36
                              Loaded: 27.78%
                              Stream Type LIVE
                              Remaining Time 0:36
                              1x
                                • Chapters
                                • descriptions off, selected
                                • captions off, selected
                                • default, selected
                                Axia ChemiSEM identifies composition on-the-fly
                                TEM videos
                                Aluminum 2099 alloy lamella characterization of Cu and Zr precipitates by APW
                                Video Player is loading.
                                Current Time 0:00
                                Duration 1:10
                                Loaded: 14.23%
                                Stream Type LIVE
                                Remaining Time 1:10
                                1x
                                  • Chapters
                                  • descriptions off, selected
                                  • captions off, selected
                                  • default, selected
                                  Nickel superalloy sample characterizingr titanium nitride nano particles by APW.
                                  Video Player is loading.
                                  Current Time 0:00
                                  Duration 1:04
                                  Loaded: 15.37%
                                  Stream Type LIVE
                                  Remaining Time 1:04
                                  1x
                                    • Chapters
                                    • descriptions off, selected
                                    • captions off, selected
                                    • en (Main), selected
                                    HSLA steel lamella characterization of Nb precipitates by Automated Particle Workflow (APW).
                                    Video Player is loading.
                                    Current Time 0:00
                                    Duration 0:19
                                    Loaded: 50.96%
                                    Stream Type LIVE
                                    Remaining Time 0:19
                                    1x
                                      • Chapters
                                      • descriptions off, selected
                                      • captions off, selected
                                      • default, selected
                                      3D EDS TEM tomography of precipitates in an AlMgSi alloy.
                                      Video Player is loading.
                                      Current Time 0:00
                                      Duration 0:53
                                      Loaded: 18.43%
                                      Stream Type LIVE
                                      Remaining Time 0:53
                                      1x
                                        • Chapters
                                        • descriptions off, selected
                                        • captions off, selected
                                        • en (Main), selected
                                        High resolution APW showing complex features in additively manufactured stainless steel.
                                        Video Player is loading.
                                        Current Time 0:00
                                        Duration 2:02
                                        Loaded: 8.16%
                                        Stream Type LIVE
                                        Remaining Time 2:02
                                        1x
                                          • Chapters
                                          • descriptions off, selected
                                          • captions off, selected
                                          • default, selected
                                          Maps and Avizo2D recordings (left and right) running side by side during an acquisition.
                                          Webinars
                                          Video Player is loading.
                                          Current Time 0:00
                                          Duration 10:03
                                          Loaded: 1.63%
                                          Stream Type LIVE
                                          Remaining Time 10:03
                                          1x
                                            • Chapters
                                            • descriptions off, selected
                                            • captions off, selected
                                            • en (Main), selected
                                            Webinar: Nanoparticle Characterization by Automated TEM.
                                            Video Player is loading.
                                            Current Time 0:00
                                            Duration 14:35
                                            Loaded: 1.13%
                                            Stream Type LIVE
                                            Remaining Time 14:35
                                            1x
                                              • Chapters
                                              • descriptions off, selected
                                              • captions off, selected
                                              • en (Main), selected
                                              Webinar: Correlative Microscopy for Aerospace and Defense Industries
                                              TEM Articles
                                              Nanoscale origins of the oriented precipitation of Ti3Al in Ti\\Al systems
                                              Hao Wu, Guohua Fan, Lin Geng, Xiping Cui, Meng Huang
                                              DOI link
                                              Effect of heat treatments on microstructural evolution of additively manufactured and wrought 17-4PH stainless steel
                                              Yu Sun, Rainer J. Hebert, Mark Aindow
                                              DOI link
                                              Coherency strains of H-phase precipitates and their influence on functional properties of nickel-titanium-hafnium shape memory alloys
                                              Behnam Amin-Ahmadi,⁎, Joseph G. Pauza, Ali Shamimi, Tom W. Duerig, Ronald D. Noebe, Aaron P. Stebner
                                              DOI link
                                              Effect of laser scan length on the microstructure of additively manufactured 17-4PH stainless steel thin-walled parts
                                              Yu Sun, Rainer J. Hebert, Mark Aindow
                                              DOI link
                                              Non-metallic inclusions in 17-4PH stainless steel parts produced by selective laser melting
                                              Yu Sun, Rainer J. Hebert, Mark Aindow
                                              DOI link
                                              FIB-SEM Articles
                                              Joachim Mayer, RWTH Aachen
                                              “Formation of White Etching Areas in SAE 52100 Bearing Steel under Rolling Contact Fatigue − Influence of Diffusible Hydrogen”
                                              M. Oezel, A. Schwedt, T. Janitzky, R. Kelley, C.Bouchet-Marquis, L. Pullan, C. Broeckmann, J. Mayer
                                              Wear, Volumes 414-415, November 2018, Pages 352-365.
                                              DOI link
                                              Philip Withers, University of Manchester
                                              “Industrial Gear Oils: Tribological Performance and Subsurface Changes”
                                              Aduragbemi Adebogun, Robert Hudson, Angela Breakspear, Chris Warrens, Ali Gholinia, Allan Matthews, Philip Withers Tribology Letters (2018) 66:65.
                                              DOI link
                                              Jun Tan, Shenyang National Laboratory for Materials Science
                                              “Insight into atmospheric pitting corrosion of carbon steel via a dual-beam FIB/SEM system associated with high-resolution TEM”
                                              Corrosion Science 152 (2019) 226–233.
                                              DOI link
                                              Yu-Lung Chiu, University of Birmingham
                                              “Micro-tensile strength of a welded turbine disc superalloy”
                                              K.M. Oluwasegun, C.Cooper, Y.L.Chiu, I.P.Jones, H.Y.Li, G.Baxter
                                              Materials Science & Engineering A 596 (2014) 229–235.
                                              DOI link
                                              Chris Pistorius, Carnegie Mellon University
                                              “Application of Plasma FIB to Analyze a Single Oxide Inclusion in Steel”
                                              D. Kumar, N.T. Nuhfer, M.E. Ferreira and P.C. Pistorius
                                              Metallurgical and Materials Transactions B, Volume 50B, June 2019, Pages 1124-1127.
                                              DOI link
                                              Applications
                                              Process control using electron microscopy
                                              Process control using electron microscopy
                                              Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
                                              Quality control and failure analysis using electron microscopy
                                              Quality control and failure analysis
                                              Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
                                              Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
                                              Fundamental Materials Research
                                              Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
                                              Aluminum mineral grain found with SEM during parts cleanliness testing
                                              Technical Cleanliness
                                              More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
                                              Style Sheet for Komodo Tabs
                                              Techniques
                                              (S)TEM Sample Preparation
                                              DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
                                              Learn more ›
                                              3D Materials Characterization
                                              Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
                                              Learn more ›
                                              Energy Dispersive Spectroscopy
                                              Energy dispersive spectroscopy (EDS) collects detailed elemental information along with electron microscopy images, providing critical compositional context for EM observations. With EDS, chemical composition can be determined from quick, holistic surface scans down to individual atoms.
                                              Learn more ›
                                              EDS Elemental Analysis
                                              Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
                                              Learn more ›
                                              3D EDS Tomography
                                              Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
                                              Learn more ›
                                              EDS Analysis with ChemiSEM Technology
                                              Energy dispersive X-ray spectroscopy for materials characterization.
                                              Learn more ›
                                              Cross-sectioning
                                              Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
                                              Learn more ›
                                              In Situ experimentation
                                              Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
                                              Learn more ›
                                              Particle analysis
                                              Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                                              Learn more ›
                                              X-Ray Photoelectron Spectroscopy
                                              X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
                                              Learn more ›
                                              Automated Particle Workflow
                                              The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
                                              Learn more ›
                                              (S)TEM Sample Preparation
                                              DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
                                              Learn more ›
                                              3D Materials Characterization
                                              Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
                                              Learn more ›
                                              Energy Dispersive Spectroscopy
                                              Energy dispersive spectroscopy (EDS) collects detailed elemental information along with electron microscopy images, providing critical compositional context for EM observations. With EDS, chemical composition can be determined from quick, holistic surface scans down to individual atoms.
                                              Learn more ›
                                              EDS Elemental Analysis
                                              Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
                                              Learn more ›
                                              3D EDS Tomography
                                              Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
                                              Learn more ›
                                              EDS Analysis with ChemiSEM Technology
                                              Energy dispersive X-ray spectroscopy for materials characterization.
                                              Learn more ›
                                              Cross-sectioning
                                              Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
                                              Learn more ›
                                              In Situ experimentation
                                              Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
                                              Learn more ›
                                              Particle analysis
                                              Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                                              Learn more ›
                                              X-Ray Photoelectron Spectroscopy
                                              X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
                                              Learn more ›
                                              Automated Particle Workflow
                                              The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
                                              Learn more ›
                                              Products
                                              Style Sheet for Instrument Cards Original
                                              STEM Sample Holder
                                              • Delivers high contrast under low voltage
                                              • Accommodates a range of materials
                                              • Includes BF, DF, and HAADF imaging modes
                                              Phenom ParticleX Steel Desktop SEM
                                              • SEM and EDS integrated
                                              • Ease of use
                                              • Sub-micrometer inclusions
                                              Talos F200S TEM
                                              • Intuitive and easy-to-use automation software
                                              • Available with Super-X EDS for rapid quantitative chemical analysis
                                              • High-throughput with simultaneous multi-signal acquisition
                                              Talos F200X TEM
                                              • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
                                              • X-FEG and X-CFEG available for the highest brightness and energy resolution
                                              • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
                                              Thermo Scientific Talos F200C transmission electron microscope (TEM)
                                              Talos F200C TEM
                                              • High-contrast and high-quality TEM and STEM imaging
                                              • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
                                              • Large pole piece gap and multiple in situ options
                                              Talos F200i TEM
                                              • Compact design with X-TWIN objective lens
                                              • Available with S-FEG, X-FEG, and X-CFEG
                                              • Flexible and fast EDS options for comprehensive elemental analysis
                                              Helios 5 HX/Helios 5 UX/Helios 5 FX DualBeam
                                              • Fully automated, high-quality, ultra-thin TEM sample preparation
                                              • High throughput, high resolution subsurface and 3D characterization
                                              • Rapid nanoprototyping capabilities
                                              Helios 5 PFIB DualBeam
                                              • Gallium-free STEM and TEM sample preparation
                                              • Multi-modal subsurface and 3D information
                                              • Next-generation 2.5 μA xenon plasma FIB column
                                              Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
                                              Scios 2 DualBeam
                                              • Full support of magnetic and non-conductive samples
                                              • High throughput subsurface and 3D characterization
                                              • Advanced ease of use and automation capabilities
                                              Thermo Scientific Apreo 2 scanning electron microscope (SEM)
                                              Apreo 2 SEM
                                              • High-performance SEM for all-round nanometer or sub-nanometer resolution
                                              • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
                                              • Excellent performance at long working distance (10 mm)
                                              Phenom Pharos G2 Desktop FEG-SEM
                                              • FEG source with 1 – 20 kV acceleration voltage range
                                              • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
                                              • Optional fully integrated EDS and SE detector
                                              Phenom ParticleX TC Desktop SEM
                                              • Versatile desktop SEM with automation software for Technical Cleanliness
                                              • Resolution <10 nm; magnification up to 200,000x
                                              • Optional SE detector
                                              Nexsa G2 XPS
                                              • Micro-focus X-ray sources
                                              • Unique multi-technique options
                                              • Dual-mode ion source for monoatomic & cluster ion depth profiling
                                              K-Alpha XPS
                                              • High resolution XPS
                                              • Fast, efficient, automated workflow
                                              • Ion source for depth profiling
                                              ESCALAB QXi XPS
                                              • High spectral resolution
                                              • Multi-technique surface analysis
                                              • Extensive sample preparation and expansion options
                                              Avizo Software
                                              Materials Science
                                              • Support for multi-data/multi-view, multi-channel, time series, very large data
                                              • Advanced multi-mode 2D/3D automatic registration
                                              • Artifact reduction algorithms
                                              Athena Software
                                              Imaging Data Management
                                              • Ensure traceability of images, data, metadata and experimental workflows
                                              • Simplify your imaging workflow​
                                              • Improve collaboration
                                              • Secure and manage data access​
                                              AutoTEM 5
                                              • Fully automated in situ S/TEM sample preparation
                                              • Support of top-down, planar and inverted geometry
                                              • Highly configurable workflow
                                              • Easy to use, intuitive user interface
                                              Thermo Scientific Maps electron microscopy software
                                              Maps Software
                                              • Acquire high-resolution images over large areas
                                              • Easily find regions of interest
                                              • Automate image acquisition process
                                              • Correlate data from different sources
                                              3D Reconstruction
                                              • Intuitive user interface, maximum employability
                                              • Intuitive fully automated user interface
                                              • Based on 'shape from shading' technology, no stage tilt required
                                              Metallurgical Sample Holder
                                              • Designed to support resin-mounted samples
                                              • Preferred solution for metallurgy and when working with inserts
                                              • Sample size up to 32 mm diameter and 30 mm height
                                              μHeater
                                              • Ultra-fast heating solution for in situ high resolution imaging
                                              • Fully integrated
                                              • Temperatures up to 1200 °C
                                              Tensile Sample Holder
                                              • Determine batch quality
                                              • Determine manufacturing consistencies
                                              • Aid the design process
                                              Velox
                                              • An experiments panel on the left side of the processing window.
                                              • Live quantitative mapping
                                              • Interactive detector layout interface for reproducible experiment control and setup
                                              Style Sheet to change H2 style to p with em-h2-header class
                                              Contact us
                                              Style Sheet for Support and Service footer
                                              Style Sheet for Fonts
                                              Style Sheet for Cards
                                              Electron microscopy services for
                                              the materials science
                                              To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.