protocol
Introduction
Since the introduction of terminal deoxynucleotidyl transferase-dUTP nick end labeling  (TUNEL) assay in 1992,1 the TUNEL assay is the most widely used in situ test for apoptosis study.2 TUNEL assay is based on the incorporation of modified dUTPs by the enzyme terminal deoxynucleotidyl transferase (TdT) at the 3’-OH ends of fragmented DNA, a hallmark as well as the ultimate determinate of apoptosis. The modifications are fluorophoresor haptens, including biotin or bromine which can be detected directly in the case of a fluorescently-modified nucleotide (i.e., fluorescein-dUTP), or indirectly with streptavidin or antibodies, if biotin-dUTP or BrdUTP are used, respectively. Often at late stages of apoptosis, adherent cells are known to detach or “pop” off. For a reliable and reproducible TUNEL imaging assay, the modified nucleotide must not only be an acceptable substrate for TdT, but the detection method must also be sensitive without bringing about any additional loss of cells from the sample.  This manual includes protocols to perform the Invitrogen Click-iT TUNEL imaging assay on adherent cells grown on coverslips (refer to the Experimental Protocol for Cells Grown on Coverslips) or a 96-well microplate (refer to Experimental Protocol for Cells Grown in 96-well Plate).
System Overview
The Invitrogen Click-iT TUNEL Alexa Fluor imaging assays utilize a dUTP modified with an alkyne, a small, bio-orthogonal functional group that enables the nucleotide to be more readily incorporated by TdT than other modified nucleotides (Figures 1–2). Detection is based on a click reaction,3-6 a copper (I) catalyzed reaction between an azide and alkyne (Figure 3).  Click chemistry fills the void when methods such as direct labeling or the use of antibodies are not efficient. The small size of the Invitrogen Alexa Fluor azide (MW ~1,000) compared to that of  an antibody (MW ~150,000) enables effortless penetration of complex samples with only mild fixation and permeabilization required. As a result, when compared to assays using other modified nucleotides, the Click-iT TUNEL imaging assay is fast (complete within 2 hours) and is able to detect a higher percentage of apoptotic cells under identical conditions (Figures 4–5). Furthermore, the Click-iT TUNEL assay allows multiplexing with surface and intracellular biomarker detection. (Table 1).
The Click-iT TUNEL Alexa Fluor imaging assay has been tested in HeLa, A549, and CHO K1 cells with a variety of reagents that induce apoptosis including staurosporine (Figure 6). The Click-iT TUNEL Alexa Fluor imaging assay contains all components needed to accurately and reliably detect apoptosis on adherent cells grown on coverslips or a 96-well microplate, and includes DNase I to generate strand breaks as a positive control.
Modified nucleotide structures 
Figure 1. Modified nucleotide structures. The alkyne and bromine modifications are significantly smaller than fluorescein.
Comparison of TdT incorporation of several modified nucleotides 
Figure 2. Comparison of TdT incorporation of several modified nucleotides. A 48-bp oligonucleotide was incubated with 30 units of TdT and an  equimolar mix of the modified nucleotide with three other nucleotides for 4 hours at room temperature. The TdT reaction products were then analyzed by gel electrophoresis, following application to a 20% TBE pre-cast gel and subsequently stained with Invitrogen SYBR Gold nucleic acid gel stain.
Comparison of TdT incorporation of several modified nucleotides 
Figure 3. Detection of apoptosis with the Click-iT® TUNEL imaging assay.
TUNEL assay comparison—percentage positives detected Figure 4. TUNEL assay comparison—percentage positives detected. HeLa cells were treated  with 0.5 µM staurosporine for 4 hours. Following fixation and permeabilization, TUNEL imaging assays were performed according to the manufacturer’s instructions. The percent positives were calculated based upon the corresponding negative control. Imaging and analysis was performed using a Thermo Fisher Scientific Cellomics ArrayScan II.
TUNEL assay time course comparison—percent positives Figure 5. TUNEL assay time course comparison—percent positives detected. HeLa cells were treated with 0.5 µM staurosporine for the time points indicated. Following fixation and permeabilization, Click-iT TUNEL imaging assays using Click-iT EdUTP or fluorescein dUTP (Promega’s DeadEnd Fluorometric TUNEL system) were performed according to the manufacturer’s instructions. The percent positives were calculated based upon the corresponding negative control. Imaging and analysis was performed using a Thermo Fisher Scientific Cellomics ArrayScan II.
Dose response of staurosporine using Click-iT® TUNEL Alexa Figure 6. Dose response of staurosporine using Click-iT TUNEL Alexa Fluor 488 imaging assay. HeLa cells were treated with staurosporine for 18 hours at final concentrations ranging from 1 nM to 1 µM. Following fixation and permeabilization, the Click-iT TUNEL imaging assay was performed.. Data points represent averages from 8 wells, the error bars show standard deviations.
Table 1. Click-iT detection reagent compatibility.
MoleculeCompatibility*
Qdot nanocrystalsUse Invitrogen Qdot nanocrystals after the Click-iT detection reaction.
Fluorescent proteinsUse Invitrogen organic dye-based reagents, such as TC-FlAsH or TC-ReAsH reagents for protein expression detection or anti-GFP rabbit or chicken antibodies after the Click-iT detection reaction.
PhalloidinClick chemistry is not compatible with phalloidin staining of F-actin. To stain the cytoskeleton, antibodies, such as anti-a-tubulin after the Click-iT detection reaction are recommended.
Organic dyes such as Alexa Fluor dyes or fluorescein isothiocyanate (FITC)Completely compatible with the Click-iT detection reaction.
TC-FlAsH or TC-ReAsH reagentsDetect the tetracysteine (TC) tag with FlAsH or ReAsH reagents before the Click-iT detection reaction.
*Some molecules or detection methods involve components that are unstable in the presence of copper catalyst used for the Click-iT detection reaction.
Materials
Table 2. Contents and storage information.
MaterialAmountConcentrationStorage*Stability
TdT reaction buffer (Component A) 11 mL
1X Solution
  • ≤–20°C
  • Desiccate
  • Protect from light
When stored as directed, kit
components are stable for at least
1 year.
EdUTP nucleotide mixture
(Component B)
100 µL50X solution
TdT (terminal deoxynucleotidyl
transferase) *recombinant*
(Component C)
6 vialsEach vial contains 34 µL enzyme at
15 U/µL in glycerol
Click-iT reaction buff er
(Component D) 
25 mL1X solution (contains Alexa Fluor 488
azide for Cat. No. C10245, Alexa Fluor
594 azide for Cat. No. C10246, Alexa
Fluor 647 azide for Cat. No. C10247)
Click-iT reaction buff er additive
(Component E)
50 mg
Not applicable
Hoechst 33342 (Component F)18 µL10 mg/mL solution in water
DNase I (deoxyribonuclease I,
Component G) 
18 µLNot applicable
DNase I buffer (Component H)
190 µL
10X solution  
*These storage conditions are appropriate when storing the entire kit upon receipt. For optimal storage of each component, see labels on individual components.
Number of assays: Sufficient material is supplied for 1 × 96-well plate or 50 coverslips based on protocols below.
Approximate fluorescence excitation/emission maxima:
  • Invitrogen Alexa Fluor 488 azide: 495/519 in nm
  • Invitrogen Alexa Fluor 594 azide: 590/615 in nm
  • Invitrogen Alexa Fluor 647 azide: 650/670 in nm
  • Hoechst 33342: 350/461 in nm, when bound to DNA.
Material Required but Not  Provided
  • 1X Phosphate buffered saline (PBS, Cat. No. 14190-144 or 14190-250)
  • 4% paraformaldehyde in PBS (fixative)
  • 0.25% Invitrogen Triton X-100 in PBS (permeabilization reagent)
  • 3% Bovine serum albumin in PBS (3% BSA in PBS), pH 7.4
  • Molecular biology grade water (DNase/RNase free)
  • 96-well plate (as recommended for the specific, automated imaging instrument)
  • 22 × 22 mm or 18 × 18 mm coverslips (for standard microscopy)
Caution
TdT reaction buffer (Component A) contains potassium cacodylate and cobalt chloride, and is harmful if swallowed. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. If swallowed seek medical advice immediately. Wear appropriate laboratory protective clothing, gloves, and eye/face protection when handling this reagent.
Hoechst 33342 (Component F) is a known mutagen. Use the dye with appropriate precautions.
Ordering Information
Catalog # Name Size Price (USD) Qty
10533073 Terminal Deoxynucleotidyl Transferase, recombinant, 3 x 500 U Each
622.65
Online Exclusive
666.00
Save 43.35 (7%)
10533065 Terminal Deoxynucleotidyl Transferase, recombinant, 500 U Each
291.65
Online Exclusive
312.00
Save 20.35 (7%)
14190144 DPBS, no calcium, no magnesium, 500 mL bottle Each
30.65
Online Exclusive
32.63
Save 1.98 (6%)
14190250 DPBS, no calcium, no magnesium, 10 x 500 mL bottle Each
263.65
Online Exclusive
285.00
Save 21.35 (7%)
18068015 DNase I, Amplification Grade Each
206.00
H1399 Hoechst 33342, trihydrochloride trihydrate, 100 mg, 100 mg Each
109.65
Online Exclusive
113.00
Save 3.35 (3%)
H3570 Hoechst 33342, trihydrochloride trihydrate, 10 mg/mL solution in water, 10 mL Each
142.65
Online Exclusive
147.00
Save 4.35 (3%)
H21492 Hoechst 33342, trihydrochloride trihydrate, FluoroPure™ grade, 100 mg, 100 mg Each
129.65
Online Exclusive
133.00
Save 3.35 (3%)
C10245 Click-iT™ TUNEL Alexa Fluor Imaging Assays for Microscopy & HCS, 1 kit Each
1,284.65
Online Exclusive
1,324.00
Save 39.35 (3%)
C10246 Click-iT™ TUNEL Alexa Fluor Imaging Assays for Microscopy & HCS, 1 kit Each
1,259.65
Online Exclusive
1,298.00
Save 38.35 (3%)
C10247 Click-iT™ TUNEL Alexa Fluor Imaging Assays for Microscopy & HCS, 1 kit Each
1,229.65
Online Exclusive
1,290.00
Save 60.35 (5%)
Protocol For Cells On Coverslips
The following protocol was developed using HeLa cells treated with 0.5 µM staurosporine for 4 hours to induce apoptosis. Cell type and treatment may influence the number of apoptotic cells detected.
1. Cell Fixation and Permeabilization
This protocol is optimized with a fixation step using 4% paraformaldehyde in PBS followed by a permeabilization step with 0.25% Triton X-100, but is amenable to other fixation and permeabilization reagents such as 70% ethanol.
  1. Remove media and wash coverslips once with PBS. Note: If there is a chance that cells maybe lost by this wash step, proceed directly to fixation (step 2) without performing the wash step.
  2. Add a sufficient volume of fixative (4% paraformaldehyde) to completely cover the coverslips.
  3. Incubate samples for 15 minutes at room temperature.
  4. Remove fixative.
  5. Add sufficient volume of the permeabilization reagent (0.25% Triton X-100 in PBS) to completely cover the coverslips.
  6. Incubate samples for 20 minutes at room temperature. Then wash twice with deionized water.
2. Preparing a Positive Control (Optional)
The DNase I generates strand breaks in the DNA to provide a positive TUNEL reaction. There is sufficient DNase I to perform ~18 positive controls.
  1. Wash coverslips with deionized or molecular biology grade water.
  2. Prepare DNase I solution according to Table 3 and mix well. Note: Do not vortex the DNase I solution as DNase I denatures with vigorous mixing.
  3. Table 3. DNase I Solution.
    Reaction ComponentsNumber of Coverslips
     1
    2
    3
    Deionized water89 µL178 µL267 µL
    DNase I buffer (Component H)10 µL20 µL30 µL
    DNase I (Component G)1 µL2 µL3 µL
    Total Volume100 µL200 µL300 µL
  4. Add 100 µL of the DNase I solution (prepared in step 2) to each coverslip and incubate for 30 minutes at room temperature.
  5. Wash coverslips once with deionized water, proceed to TdT Reaction.
3. TdT Reaction
This protocol uses 100 µL of the TdT reaction cocktail per coverslip. It is important toequilibrate cells to maximize efficiency of the TdT reaction.
  1. Add 100 µL TdT reaction buffer (Component A) to each coverslip and allow the solution to spread completely over the surface. 
  2. Incubate coverslips for 10 minutes at room temperature.
  3. Remove TdT reaction buffer.
  4. Prepare the TdT reaction cocktail according to Table 4.
  5. Table 4. TdT Reaction Cocktails.
    Reaction ComponentsNumber of Coverslips
     124510
    TdT reaction buffer (Component A)94 µL188 µL376 µL470 µL940 µL
    EdUTP (Component B)2 µL4 µL8 µL10 µL20 µL
    TdT* (Component C) 4 µL8 µL16 µL20 µL 40 µL
    Total Volume100 µL200 µL400 µL500 µL1 mL
  6. Add 100 µL of the TdT reaction cocktail (prepared in step 4) to each coverslip and allow the solution to spread completely over the surface.
  7. Incubate coverslips for 60 minutes at 37°C. Use a humidified chamber to protect against evaporation. Remove the reaction cocktail.  Note: This reaction can be performed overnight at room temperature.
  8. Wash coverslips twice with 3% BSA in PBS for 2 minutes each.
4. Click-iT Reaction
  1. Prepare the Click-iT reaction buffer additive (Component E) by adding 625 µL of deionized water to the Component E vial. After use, aliquot and store any remaining solution at =–20°C.  When stored as directed the stock solution is stable for up to 1 year.
  2. Prepare the Click-iT reaction cocktail according to Table 5 and mix well by vortexing. Note: Use the Click-iT reaction cocktail within 15 minutes of preparation.
  3. Immediately add 100 µL of the Click-iT reaction cocktail (prepared in step 2) to each coverslip and allow the solution to spread completely over the surface.
  4. Table 5. Click-iT Reaction Cocktails.
    Reaction ComponentsNumber of Coverslips
     124510
    Click-iT reaction buffer (Component D)97.5 µL195 µL 390 µL487.5 µL975 µL
    Click-iT reaction buffer additive (prepared in step 4.2)2.5 µL5 µL10 µL12.5 µL25 µL
    Total Volume100 µL200 µL400 µL500 µL1 mL
  5. Incubate coverslips for 30 minutes at room temperature, protected from light.
  6. Remove the Click-iT reaction cocktail and wash each coverslip with 3% BSA in 1X PBS for 5 minutes.
  7. For antibody staining, proceed to section 5, or for DNA staining, proceed to section 6. If no additional staining is desired, proceed to Imaging and Analysis.
5. Antibody Detection (Optional)
  1. If required, block the coverslips or wells with 3% BSA in 1X PBS for the recommended time, protected from light. Remove the blocking solution.
  2. Prepare and add the primary antibody solution as recommended by the manufacturer.
  3. Incubate for the recommended time and temperature, protected from light. Remove the primary antibody solution.
  4. Wash each coverslip or well twice with 3% BSA in PBS. Remove the wash solution.
  5. Prepare and add the secondary antibody solution as recommended by the manufacturer.
  6. Incubate for the recommended time and temperature, protected from light. Remove the secondary antibody solution.
  7. Wash each coverslip or well twice with 3% BSA in PBS. Remove the wash solution.
For DNA staining, proceed to section 6. If no additional staining is desired, proceed to Imaging and Analysis.
6. DNA Staining
  1. Dilute Hoechst 33342 (Component F) 1:5,000 in PBS to obtain a 1X Hoechst 33342 solution.
  2. Add 100 µL 1X Hoechst 33342 solution per coverslip or well and incubate for 15 minutes at room temperature, protected from light. Remove the Hoechst 33342 solution.
  3. Wash each coverslip or well twice with PBS. Remove the wash solution.
Imaging and Analysis
The Click-iT TUNEL stained cells are compatible with all methods of slide preparation including wet mount or prepared mounting media. See Table 6 for the approximate fluorescence excitation/emission maxima for Alexa Fluor dyes and Hoechst 33342 bound to DNA.
Table 6. Approximate Fluorescence Excitation/emission Maxima.
Fluorophore Excitation (nm)Emission (nm)
Alexa Fluor 488495519
Alexa Fluor 594590615
Alexa Fluor 647650670
Hoechst 33342 bound to DNA350460
Protocol For Cells in 96-Well Plates
The following protocol was developed using HeLa cells treated with 0.5 µM staurosporine for 4 hours to induce apoptosis. Cell type and treatment may influence the number of apoptotic cells detected.
7.  Cell Fixation and Permeabilization
This protocol is optimized with a fixation step using 4% paraformaldehyde in PBS followed by a permeabilization step with 0.25% Triton X-100, but is amenable to other fixation and permeabilization reagents such as 70% ethanol.
  1. Remove media and wash wells once with PBS. Note: If there is a chance that cells maybe lost by this wash step, proceed directly to fixation (step 2) without performing the wash step.
  2. Add 100 µL fixative (4% paraformaldehyde) to completely cover each well.
  3. Incubate plates for 15 minutes at room temperature.
  4. Remove fixative.
  5. Add 100 µL of the permeabilization reagent (0.25% Triton X-100 in PBS) to completely cover each well.
  6. Incubate plates for 20 minutes at room temperature.
  7. Wash twice with deionized water.
8.  Preparing a Positive Control (Optional)
The DNase I generates strand breaks in the DNA to provide a positive TUNEL reaction. There is sufficient DNase I to perform ~18 positive controls.
  1. Wash the wells with deionized or molecular biology grade water.
  2. Prepare DNase I solution according to Table 7 and mix well. Note: Do not vortex the DNase I solution as DNase I denatures with vigorous mixing.
  3. Table 7. DNase I Solution.
    Reaction ComponentsNumber of Wells
     1
    2
    4
    5
    8
    Deionized water89 µL178 µL356 µL445 µL712 µL
    DNase I buffer (Component H)10 µL20 µL40 µL50 µL80 µL
    DNase I (Component G)1 µL2 µL4 µL5 µL8 µL
    Total Volume100 µL200 µL400 µL500 µL800 µL
  4. Add 100 µL of the DNase I solution (prepared in step 2) to each coverslip and incubate for 30 minutes at room temperature.
  5. Wash coverslips once with deionized water, proceed to TdT Reaction.
9.  TdT Reaction
This protocol uses 50 µL of the TdT reaction cocktail per well. It is important to equilibrate cells to maximize efficiency of the TdT reaction.
  1. Add 50 µL TdT reaction buffer (Component A) to each well and allow the solution to spread completely over the surface.
  2. Incubate plates for 10 minutes at room temperature.
  3. Remove TdT reaction buffer.
  4. Prepare the TdT reaction cocktail according to Table 8.
  5. Table 8. TdT Reaction Cocktails.
    Reaction ComponentsNumber of Wells
     1812100 
    TdT reaction buffer (Component A)47 µL376 µL564 µL4.7 mL 
    EdUTP (Component B)1 µL8 µL12 µL100 µL 
    TdT* (Component C) 2 µL16 µL24 µL200 µL 
    Total Volume50 µL400 µL600 µL5 mL 
  6. Add 50 µL of the TdT reaction cocktail (prepared in step 4) to each well and allow the solution to spread completely over the well.
  7. Incubate plates for 60 minutes at 37°C. Remove the reaction cocktail.
  8. Wash wells twice with 3% BSA in PBS for 2 minutes each.
10.  Click-iT Reaction
  1. Prepare the Click-iT reaction buffer additive (Component E) by adding 625 µL of deionized water to the Component E vial. After use, aliquot and store any remaining solution at =–20°C.  When stored as directed the stock solution is stable for up to 1 year.
  2. Prepare the Click-iT reaction cocktail according to Table 9 and mix well by vortexing. Note: Use the Click-iT reaction cocktail within 15 minutes of preparation.
  3. Table 9. Click-iT Reaction Cocktails.
    Reaction ComponentsNumber of Coverslips
     1812100 
    Click-iT reaction buffer (Component D)97.5 µL780 µL 1.17 mL9.75 mL 
    Click-iT reaction buffer additive (prepared in step 10.1)2.5 µL20 µL30 µL250 µL 
    Total Volume100 µL800 µL1.2 mL10 mL 
  4. Immediately add 50 µL of the Invitrogen Click-iT reaction cocktail (prepared in step 10.2) to each well and allow the solution to spread completely over the surface.
  5. Incubate plates for 30 minutes at room temperature, protected from light. Remove the reaction cocktail
  6. Wash each coverslip with 3% BSA in 1X PBS for 5 minutes.
  7. Optional: Stain with antibodies (section 5) or other stains such as DNA staining for staining nuclei with Hoechst 33342 (section 6).
  8. Image plate using filters appropriate for the Alexa Fluor dye included in the Click-iT TUNEL imaging assay (Table 6) and any other fluorophores used.
References
  1.    J Cell Biol 119, 493 (1992)
  2.    J Surg Res 139, 143 (2007)
  3.    ChemBioChem 4, 1147 (2003)
  4.    J Am Chem Soc 125, 3192 (2003)
  5.    Angew Chem Int Ed Engl 41, 2596 (2002)
  6.    Angew Chem Int Ed Engl 40, 2004 (2001). 
MP10245      14–Nov–2008