1x1 image pixel for data collection
DEPC-Treated Water
DEPC-Treated Water
Invitrogen™

DEPC-Treated Water

Ambion DEPC-treated water is certified nuclease-free and supplied in five bottles containing 100 mL each. DEPC-treated water is autoclaved pre-Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 x 100 mL
1 x 500 mL
10 x 50 mL
5 x 100 mL
1 x 1000 mL
4 x 1000 mL
Catalog number 4387937
Price (USD)
249.65
Online Exclusive
264.00
Save 14.35 (5%)
Each
In stock
Add to cart
Quantity:
4 x 1000 mL
Recurring order eligible. Learn more »
Price (USD)
249.65
Online Exclusive
264.00
Save 14.35 (5%)
Each
Add to cart
Ask our AI about this Product

Ambion DEPC-treated water is certified nuclease-free and supplied in five bottles containing 100 mL each. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineAmbion™
Purification MethodAutoclaved, Membrane-Filtered
Recommended StorageStore at room temperature.
Treatment(s)DEPC-Treated
GradeMolecular Biology
Quantity4 x 1000 mL
pH6 to 8
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How can DEPC-treated water be prepared?

DEPC-treated (ribonuclease free) water can be prepared as decribed below:

Add 1 mL DEPC per L of H2O

Shake until DEPC has dissolved in the water

Let sit for 1 h

Autoclave

Buffers containing Tris cannot be DEPC-treated (Tris inactivates DEPC by binding to amino groups). To prepare a Tris buffer, the water should be DEPC-treated and autoclaved before adding Tris. After addition of the Tris, the solution should be autoclaved again.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2505057Certificate of AnalysisJul 10, 2025AM9906
2505055Certificate of AnalysisJun 27, 2025AM9915G
2505056Certificate of AnalysisJun 27, 2025AM9906
2003032Certificate of AnalysisJun 27, 2025AM9915G
2503053Certificate of AnalysisJun 27, 2025AM9915G
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection