Search Thermo Fisher Scientific
Our lines of basic, standard, advanced, and specialty ceramic hot plates offer fast heat up times and reliable performance. Ceramic surfaces provide benefits such as resistance to chemicals and corrosion and the ability to handle higher temperatures than aluminum surfaces. Most models feature white surfaces for optimal sample viewing.
Find the hotplate to meet your needs using our hot plate selection guide ›
Cimarec Basic Hotplates | ||
---|---|---|
![]() | ||
Reach maximum temperature in under 8 minutes with Thermo Scientific Cimarec Basic Hotplates. A sleek, rugged design and construction reduces the chance of top plate breakage and promotes durability, for economical high performance. | ||
Part no. | HP194515 | HP194825 |
Plate shape | Square | Square |
Plate material | Ceramic | Ceramic |
Plate size | 4.25 x 4.25 in. | 7.25 x 7.25 in. |
Voltage | 120V | 120V |
Temperature Range | 150° to 538°C | 150° to 538°C |
Cimarec+ Hotplate Series | ||||||
---|---|---|---|---|---|---|
![]() | ||||||
Our Thermo Scientific Cimarec+ hotplate series offers exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations. | ||||||
Part no. | HP88854100 | HP88854105 | HP88857100 | HP88857105 | HP88850100 | HP88850105 |
Plate shape | Square | Square | Square | Square | Square | Square |
Plate material | Ceramic | Ceramic | Ceramic | Ceramic | Ceramic | Ceramic |
Plate size | 4.25 x 4.25 in. | 4.25 x 4.25 in. | 7.25 x 7.25 in. | 7.25 x 7.25 in. | 10.25 x 10.25 in. | 10.25 x 10.25 in. |
Voltage | 100-120V | 230V | 100-120V | 230V | 100-120V | 230V |
Max. Temperature | 540°C | 540°C | 540°C | 540°C | 540°C | 540°C |
RT2 Digital Hotplate | ||
---|---|---|
![]() | ||
Increase your confidence in safety and accuracy with the sleek, low-profile Thermo Scientific RT2 Digital Hotplate, which offers precise digital control with 0.1°C resolution and maximum temperature of 350°C. Experience fast heat-up time and an excellent heat transfer rate thanks to the tightly integrated structure of the heater and a ceramic-coated aluminum alloy top plate which provides excellent temperature uniformity and corrosion resistance while allowing for optimal monitoring of color changes in samples or media during operation. | ||
Part no. | 88880001 | 88880002 |
Plate shape | Round | Round |
Plate material | Ceramic coated aluminum | Ceramic coated aluminum |
Plate size | 5.5 in. diam. | 5.5 in. diam. |
Voltage | 230V | 120V |
Temperature Range | 50° to 350°C | 50° to 350°C |
5.5 in. diam. |
SuperNuova+ Hotplate Series | ||||
---|---|---|---|---|
![]() | ||||
Our new Thermo Scientific SuperNuova+ hotplate series provides reliable performance, dependable safety and simple operation. Units are available in two sizes, and provide optimized controls and settings for applications that demand advanced precision. | ||||
Part no. | HP88857190 | HP88857195 | HP88850190 | HP88850195 |
Plate shape | Square | Square | Square | Square |
Plate material | Ceramic | Ceramic | Ceramic | Ceramic |
Plate size | 7.25 x 7.25 in. | 7.25 x 7.25 in. | 10.25 x 10.25 in. | 10.25 x 10.25 in. |
Voltage | 100-120V | 230V | 100-120V | 230V |
Max Temperature | 450°C | 450°C | 450°C | 450°C |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.