10 to 15 Liter Carboys
The classic 10L (2.5ga) Nalgene carboy is the perfect size for many labs. Small and portable enough to find its way onto benchtops throughout the lab, yet roomy enough to get you through a good portion of the day and rugged enough to contain your most valuable solutions wherever your work may take you. Choose from a wide variety of shapes and plastic materials to find the carboy that best meets your application needs. 
  • Polypropylene (PP), polypropylene copolymer (PPCO), high density polyethylene (HDPE) and low density polyethylene (LDPE) carboys are chemically compatible with a wide range of lab reagents including most acids, bases, alcohols, aldehydes, esters, and aliphatic hydrocarbons. Choose fluorinated high density polyethylene (FLPE) for enhanced compatibility with organic solvents. 
  • Polypropylene, polypropylene copolymer and polycarbonate (PC) materials are autoclavable.
  • The polyethylenes and polycarbonate are ideal choices for low-temperature applications.
For chemical compatibility ratings by specific chemical and application temperature, consult the  Nalgene Chemical Resistance Guide, or request a copy of the printed wall chart. Nalgene carboys are made only from high-quality laboratory-grade resin materials that comply with a number of regulatory specifications including: USP Class VI, EP monographs, EU food-contact directives, CONEG, RoHS, CA Prop 65, SARA Title III Sec. 313, and 21 CFR pt 177 for food contact. For regulatory claims  and certifications applicable to specific Nalgene carboys, please contact  Nalgene Regulatory Support.
For help selecting the Thermo Scientific Nalgene carboy that's perfect for your lab applications, use our online selection guide or for personal assistance, contact the  Nalgene technical support team.
Narrow-Mouth Round Carboy with Spigot (2319-0020, 8319-0020 APAC)Narrow-Mouth Round Carboy (2250-0020, 8250-series in APAC)Wide-Mouth Round Carboy (2235-0020)Narrow-Mouth Round Carboy with Tubulation Port (2301-0020)Rectangular Carboy with Spigot (2321-0020)Rectangular Carboy (2212-0020)
Dispensing lab water and reagents.  Compatible with sterilization by autoclavingLiquid storage and shipping.  Applications where autoclaving is required.Liquid storage where autoclaving is required. Wide  mouth allows easy access for filling and cleaningFor gravity fed liquid dispensing through tubing.  Applications where autoclaving is required.Dispensing lab water and reagents.  Excellent choice for space efficient reagent banks. AutoclavableSpace-efficient storage for large volume lab solutions where autoclaving is required
PP with white PP closure/TPE gasket, PP spigotPP with white PP closure/TPE gasketPPCO with white PP closurePP with white PP closure/TPE gasketPPCO with white PP closure, PP spigotPPCO with white PP closure
RoundRoundRoundRoundRectangular Rectangular 
Milky white translucentMilky white translucentMilky white translucentMilky white translucentMilky white translucentMilky white translucent
10L (2.5ga)10L (2.5ga)10L (2.5ga)10L (2.5ga)9L (2ga)9L (2ga)
Narrow mouth Narrow mouth Wide mouthNarrow mouth Wide mouthWide mouth
SpigotNo drainNo drainTubing adapter portSpigotNo drain
Leakproof*Leakproof*Leakproof*Leakproof*Leakproof*Leakproof*
Temp range 0°C to 135°CTemp range 0°C to 135°CTemp range -40°C to 121°CTemp range 0°C to 135°CTemp range -40°C to 121°CTemp range -40°C to 121°C
AutoclavableAutoclavableAutoclavableAutoclavableAutoclavableAutoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesStainless steel handle and 20L w/molded-in gripStainless steel handle and 20L w/molded-in grip
Complies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food use
BPA-freeBPA-freeBPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Carboy with spigot (2319-0020, 8319-0020 APAC)Narrow-Mouth Round Carboy (2250-0020, 8250-Series in APAC)Wide-Mouth Round Carboy (2235-0020)Narrow-Mouth Round Carboy with Tubulation Port (2301-0020)Rectangular Carboy with Spigot (2321-0020)Rectangular Carboy (2212-0020)
Acids, dilute or weakEEEEEE
Acids^ strong, concentratedGGGGGG
Alcohols, aliphaticEEEEEE
AldehydesGGGGGG
Bases/alkaliEEEEEE
EstersGGGGGG
Hydrocarbons, aliphaticGGGGGG
Hydrocarbons, aromaticNNNNNN
Hydrocarbons, halogenated NNNNNN
Ketones, aromaticNNNNNN
Oxidizing agents, strongFFFFFF
For a more detailed chemical resistance guide by chemical and application temperature, please refer to  Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
Footnote:
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Specialty PP and PPCO Carboys
Rectangular Carboy with Spigot
(2320-0020)
Rectangular Carboy (2211-0020)Amber Carboy
(2256-7020)
Lowboy with Spigot, HDPE
(2323-0015)
Dispensing lab water and reagents. Excellent choice for space efficient reagent banks.Space-efficient storage  for large volume lab solutions. Storing and transporting large volume light sensitive solutionsLow profile design fits on shelves with tight head room like inside refrigerators or over the sink. Can stack 3 high for a reagent bank. 
HDPE with PP spigot and white PP closureHDPE with white PP  closureAmber HDPE with amber PP closureHDPE lowboy with PP closure and spigot
Rectangular Rectangular RoundLowboy design
Milky white translucentMilky white translucentOpaque amber
Milky white translucent
9L (2ga) 9L (2ga) 10L (2.5ga)15L (4ga)
Wide mouthWide mouthNarrow mouth Narrow mouth 
SpigotNo drainNo drainSpigot
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range -100°C to 120°CTemp range -100°C to 120°CTemp range -100°C to 120°CTemp range -100°C to 120°C
Not autoclavableNot autoclavableNot autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallonsNo graduationsNo graduations
Stainless steel handleStainless steel handle and 20L w/molded-in gripMolded-in shoulder handlesMolded-in handle
Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1
BPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Rectangular Carboy with Spigot 
(2320-0020)
Rectangular Carboy (2211-0020)Amber Carboy 
(2256-7020)
Lowboy with Spigot, HDPE 
(2323-0015)
Acids, dilute or weakEEEE
Acids^ strong, concentratedGGGG
Alcohols, aliphaticEEEE
AldehydesGGGG
Bases/alkaliEEEE
EstersGGGG
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot
(2318-series,  8318-series in APAC)
Narrow-Mouth Round Carboy
(2210-series, 8210-series in APAC)
Narrow-Mouth Round Carboy with Tubulation Port
(2302-0020)
Wide-Mouth Round Carboy
(2234-series)
Classic DI water and lab reagent dispenserClassic DI water and reagent reservoirFor gravity fed liquid dispensing through tubingWide mouth reagent reservoir provides easy access and cleaning
LDPE carboy with white PP closure and PP spigotLDPE carboy with white PP closureLDPE carboy with white PP closureLDPE carboy with white PP closure
RoundRoundRoundRound
Milky white translucentMilky white translucentMilky white translucentMilky white translucent
10L (2.5ga)10L (2.5ga) and 15L (3.75ga)10L (2.5ga)10L (2.5ga) and 15L (4ga)
Narrow mouth Narrow mouth Narrow mouth Wide mouth
SpigotNo drainTubing adapter portNo drain
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Not autoclavableNot autoclavableNot autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handles
Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1
BPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Carboy with Spigot 
(2318-Series, 8318-Series in APAC)
Narrow-Mouth Round Carboy 
(2210-series, 8210-series in APAC)
Narrow-Mouth Round Carboy with Tubulation Port
(2302-0020)
Wide-Mouth Round Carboy
(2234-series)
Acids, dilute or weakEEEE
Acids^ strong, concentratedGGGG
Alcohols, aliphaticEEEE
AldehydesGGGG
Bases/alkaliEEEE
EstersGGGG
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Clearboy with spigot
(2317-0020)
Narrow-Mouth Round Clearboy Carboy
(2251-0020)
Wide-Mouth Rectangular Clearboy with Spigot
(2322-0020)
Wide-Mouth Rectangular Clearboy
(DS2213-0020)
Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired.  Compatible with sterilization by autoclavingMixing, storing and shipping lab media and non-aggressive aqueous  reagents where a clear view of contents and autoclavability is desired.  Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired.  Rectangular shape optimized shelf space. AutoclavableStoring lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimizes shelf space. Autoclavable
PC with white PP closure/TPE gasket and PP spigotPC with white PP closure/TPE gasket PC with white PP closure and PP spigotPC with white PP closure 
RoundRoundRectangularRectangular
ClearClearClearClear
10L (2.5ga)10L (2.5ga)9L (2ga) 9L (2ga) 
Narrow mouth Narrow mouth Wide mouthWide mouth
SpigotNo drainSpigotNo drain
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range -135°C to 135°CTemp range -135°C to 135°CTemp range -135°C to 135°CTemp range -135°C to 135°C
AutoclavableAutoclavableAutoclavableAutoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
No handlesNo handlesStainless steel handleStainless steel handle
Complies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food use
Contains BPAContains BPAContains BPAContains BPA
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Clearboy with spigot
(2317-0020)
Narrow-Mouth Round Clearboy Carboy
(2251-0020)
Wide-Mouth Rectangular Clearboy with Spigot
(2322-0020)
Wide-Mouth Rectangular Clearboy 
(DS2213-0020)
Acids, dilute or weakEEEE
Acids^ strong, concentratedNNNN
Alcohols, aliphaticGGGG
AldehydesFFFF
Bases/alkaliNNNN
EstersNNNN
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Fluorinated Carboy (2097-0020)Fluorinated Carboy with Spigot (DS2327-0020)
Storage for  solvents and other organic solutions Space-efficient dispensing for non-flammable solvents and other organic solutions 
FLPE with white PP closureFLPE with white PP  closure and fluoropolymer spigot
RoundRectangular 
Milky white translucent, yellowish tint due to fluorination processMilky white translucent, yellowish tint due to fluorination process
10L (2.5ga) - 20L (5ga) 9L (2ga) - 20L (5ga) 
Narrow mouthWide mouth
No drainSpigot
Leakproof*Leakproof*
Temp range -100°C to 120°CTemp range -100°C to 120°C
Not autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handleStainless steel handle
Complies with Part 21 CFR 177.1615 for food use1Complies with Part 21 CFR 177.1615 for food use1
BPA-freeBPA-free
Phthalate-freePhthalate-free
General chemical compatibility
 Fluorinated Carboy
(2097-0020)
Fluorinated Carboy with Spigot
(DS2327-0020)
Acids, dilute or weakEE
Acids^ strong, concentratedGG
Alcohols, aliphaticEE
AldehydesGG
Bases/alkaliFF
EstersGG
Hydrocarbons, aliphaticEE
Hydrocarbons, aromaticEE
Hydrocarbons, halogenated GG
Ketones, aromaticGG
Oxidizing agents, strongFF
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot (2319-0020, 8319-0020 APAC)Narrow-Mouth Round Carboy (2250-0020, 8250-series in APAC)Wide-Mouth Round Carboy (2235-0020)Narrow-Mouth Round Carboy with Tubulation Port (2301-0020)Rectangular Carboy with Spigot (2321-0020)Rectangular Carboy (2212-0020)
Dispensing lab water and reagents.  Compatible with sterilization by autoclavingLiquid storage and shipping.  Applications where autoclaving is required.Liquid storage where autoclaving is required. Wide  mouth allows easy access for filling and cleaningFor gravity fed liquid dispensing through tubing.  Applications where autoclaving is required.Dispensing lab water and reagents.  Excellent choice for space efficient reagent banks. AutoclavableSpace-efficient storage for large volume lab solutions where autoclaving is required
PP with white PP closure/TPE gasket, PP spigotPP with white PP closure/TPE gasketPPCO with white PP closurePP with white PP closure/TPE gasketPPCO with white PP closure, PP spigotPPCO with white PP closure
RoundRoundRoundRoundRectangular Rectangular 
Milky white translucentMilky white translucentMilky white translucentMilky white translucentMilky white translucentMilky white translucent
10L (2.5ga)10L (2.5ga)10L (2.5ga)10L (2.5ga)9L (2ga)9L (2ga)
Narrow mouth Narrow mouth Wide mouthNarrow mouth Wide mouthWide mouth
SpigotNo drainNo drainTubing adapter portSpigotNo drain
Leakproof*Leakproof*Leakproof*Leakproof*Leakproof*Leakproof*
Temp range 0°C to 135°CTemp range 0°C to 135°CTemp range -40°C to 121°CTemp range 0°C to 135°CTemp range -40°C to 121°CTemp range -40°C to 121°C
AutoclavableAutoclavableAutoclavableAutoclavableAutoclavableAutoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesStainless steel handle and 20L w/molded-in gripStainless steel handle and 20L w/molded-in grip
Complies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food useComplies with Part 21 CFR 177.1520 for food use
BPA-freeBPA-freeBPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Carboy with spigot (2319-0020, 8319-0020 APAC)Narrow-Mouth Round Carboy (2250-0020, 8250-Series in APAC)Wide-Mouth Round Carboy (2235-0020)Narrow-Mouth Round Carboy with Tubulation Port (2301-0020)Rectangular Carboy with Spigot (2321-0020)Rectangular Carboy (2212-0020)
Acids, dilute or weakEEEEEE
Acids^ strong, concentratedGGGGGG
Alcohols, aliphaticEEEEEE
AldehydesGGGGGG
Bases/alkaliEEEEEE
EstersGGGGGG
Hydrocarbons, aliphaticGGGGGG
Hydrocarbons, aromaticNNNNNN
Hydrocarbons, halogenated NNNNNN
Ketones, aromaticNNNNNN
Oxidizing agents, strongFFFFFF
For a more detailed chemical resistance guide by chemical and application temperature, please refer to  Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
Footnote:
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Specialty PP and PPCO Carboys
Rectangular Carboy with Spigot
(2320-0020)
Rectangular Carboy (2211-0020)Amber Carboy
(2256-7020)
Lowboy with Spigot, HDPE
(2323-0015)
Dispensing lab water and reagents. Excellent choice for space efficient reagent banks.Space-efficient storage  for large volume lab solutions. Storing and transporting large volume light sensitive solutionsLow profile design fits on shelves with tight head room like inside refrigerators or over the sink. Can stack 3 high for a reagent bank. 
HDPE with PP spigot and white PP closureHDPE with white PP  closureAmber HDPE with amber PP closureHDPE lowboy with PP closure and spigot
Rectangular Rectangular RoundLowboy design
Milky white translucentMilky white translucentOpaque amber
Milky white translucent
9L (2ga) 9L (2ga) 10L (2.5ga)15L (4ga)
Wide mouthWide mouthNarrow mouth Narrow mouth 
SpigotNo drainNo drainSpigot
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range -100°C to 120°CTemp range -100°C to 120°CTemp range -100°C to 120°CTemp range -100°C to 120°C
Not autoclavableNot autoclavableNot autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallonsNo graduationsNo graduations
Stainless steel handleStainless steel handle and 20L w/molded-in gripMolded-in shoulder handlesMolded-in handle
Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1
BPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Rectangular Carboy with Spigot 
(2320-0020)
Rectangular Carboy (2211-0020)Amber Carboy 
(2256-7020)
Lowboy with Spigot, HDPE 
(2323-0015)
Acids, dilute or weakEEEE
Acids^ strong, concentratedGGGG
Alcohols, aliphaticEEEE
AldehydesGGGG
Bases/alkaliEEEE
EstersGGGG
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot
(2318-series,  8318-series in APAC)
Narrow-Mouth Round Carboy
(2210-series, 8210-series in APAC)
Narrow-Mouth Round Carboy with Tubulation Port
(2302-0020)
Wide-Mouth Round Carboy
(2234-series)
Classic DI water and lab reagent dispenserClassic DI water and reagent reservoirFor gravity fed liquid dispensing through tubingWide mouth reagent reservoir provides easy access and cleaning
LDPE carboy with white PP closure and PP spigotLDPE carboy with white PP closureLDPE carboy with white PP closureLDPE carboy with white PP closure
RoundRoundRoundRound
Milky white translucentMilky white translucentMilky white translucentMilky white translucent
10L (2.5ga)10L (2.5ga) and 15L (3.75ga)10L (2.5ga)10L (2.5ga) and 15L (4ga)
Narrow mouth Narrow mouth Narrow mouth Wide mouth
SpigotNo drainTubing adapter portNo drain
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Temp range
-76°C to 70°C
Not autoclavableNot autoclavableNot autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handlesMolded-in shoulder handles
Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1Complies with Part 21 CFR 177.1520 for food use1
BPA-freeBPA-freeBPA-freeBPA-free
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Carboy with Spigot 
(2318-Series, 8318-Series in APAC)
Narrow-Mouth Round Carboy 
(2210-series, 8210-series in APAC)
Narrow-Mouth Round Carboy with Tubulation Port
(2302-0020)
Wide-Mouth Round Carboy
(2234-series)
Acids, dilute or weakEEEE
Acids^ strong, concentratedGGGG
Alcohols, aliphaticEEEE
AldehydesGGGG
Bases/alkaliEEEE
EstersGGGG
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Clearboy with spigot
(2317-0020)
Narrow-Mouth Round Clearboy Carboy
(2251-0020)
Wide-Mouth Rectangular Clearboy with Spigot
(2322-0020)
Wide-Mouth Rectangular Clearboy
(DS2213-0020)
Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired.  Compatible with sterilization by autoclavingMixing, storing and shipping lab media and non-aggressive aqueous  reagents where a clear view of contents and autoclavability is desired.  Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired.  Rectangular shape optimized shelf space. AutoclavableStoring lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimizes shelf space. Autoclavable
PC with white PP closure/TPE gasket and PP spigotPC with white PP closure/TPE gasket PC with white PP closure and PP spigotPC with white PP closure 
RoundRoundRectangularRectangular
ClearClearClearClear
10L (2.5ga)10L (2.5ga)9L (2ga) 9L (2ga) 
Narrow mouth Narrow mouth Wide mouthWide mouth
SpigotNo drainSpigotNo drain
Leakproof*Leakproof*Leakproof*Leakproof*
Temp range -135°C to 135°CTemp range -135°C to 135°CTemp range -135°C to 135°CTemp range -135°C to 135°C
AutoclavableAutoclavableAutoclavableAutoclavable
Graduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallonsGraduated in liters and gallons
No handlesNo handlesStainless steel handleStainless steel handle
Complies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food useComplies with Part 21 CFR 177.1580 for food use
Contains BPAContains BPAContains BPAContains BPA
Phthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Narrow-Mouth Round Clearboy with spigot
(2317-0020)
Narrow-Mouth Round Clearboy Carboy
(2251-0020)
Wide-Mouth Rectangular Clearboy with Spigot
(2322-0020)
Wide-Mouth Rectangular Clearboy 
(DS2213-0020)
Acids, dilute or weakEEEE
Acids^ strong, concentratedNNNN
Alcohols, aliphaticGGGG
AldehydesFFFF
Bases/alkaliNNNN
EstersNNNN
Hydrocarbons, aliphaticGGGG
Hydrocarbons, aromaticNNNN
Hydrocarbons, halogenated NNNN
Ketones, aromaticNNNN
Oxidizing agents, strongFFFF
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Fluorinated Carboy (2097-0020)Fluorinated Carboy with Spigot (DS2327-0020)
Storage for  solvents and other organic solutions Space-efficient dispensing for non-flammable solvents and other organic solutions 
FLPE with white PP closureFLPE with white PP  closure and fluoropolymer spigot
RoundRectangular 
Milky white translucent, yellowish tint due to fluorination processMilky white translucent, yellowish tint due to fluorination process
10L (2.5ga) - 20L (5ga) 9L (2ga) - 20L (5ga) 
Narrow mouthWide mouth
No drainSpigot
Leakproof*Leakproof*
Temp range -100°C to 120°CTemp range -100°C to 120°C
Not autoclavableNot autoclavable
Graduated in liters and gallonsGraduated in liters and gallons
Molded-in shoulder handleStainless steel handle
Complies with Part 21 CFR 177.1615 for food use1Complies with Part 21 CFR 177.1615 for food use1
BPA-freeBPA-free
Phthalate-freePhthalate-free
General chemical compatibility
 Fluorinated Carboy
(2097-0020)
Fluorinated Carboy with Spigot
(DS2327-0020)
Acids, dilute or weakEE
Acids^ strong, concentratedGG
Alcohols, aliphaticEE
AldehydesGG
Bases/alkaliFF
EstersGG
Hydrocarbons, aliphaticEE
Hydrocarbons, aromaticEE
Hydrocarbons, halogenated GG
Ketones, aromaticGG
Oxidizing agents, strongFF
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
 E No damage after 30 days constant exposure at 20oC. 
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Technical support
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
  technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to
  techsupport.labproducts.eu@thermofisher.com.
Regulatory Support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at  RocRegSupport@thermofisher.com
Resources
Additional resources
  • Break the Glass Habit Brochure
  • Bottle and Carboy Selection Guide
  • Plastic Properties Reference Magnet
  • Plastic Labware Chemical Resistance Wall Poster
Request resources
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.