Search Thermo Fisher Scientific
Nalgene 20L (5ga) and 25L (6.5ga) Nalgene carboys go the extra mile containing your higher-volume lab solutions. Their rugged construction will hold up to the most rigorous lab and field applications. Made only from the highest-quality laboratory,
For chemical compatibility ratings by specific chemical and application temperature, consult the Nalgene Chemical Resistance Guide, or request a copy of the printed wall chart. Nalgene carboys are made only from high-quality laboratory-grade resin materials that comply with a number of regulatory specifications including: USP Class VI, EP monographs, EU food-contact directives, CONEG, RoHS, CA Prop 65, SARA Title III Sec. 313, and 21 CFR pt 177 for food contact. For regulatory claims and certifications applicable to specific Nalgene carboys, please contact Nalgene Regulatory Support.
For help selecting the Thermo Scientific Nalgene carboy that's perfect for your lab applications, use our online selection guide or for personal assistance, contact the Nalgene technical support team.
Narrow-Mouth Round Carboy with Spigot (2319-0050, 8319-0050 APAC) | Narrow-Mouth Round Carboy (2250-0050, 8250-0050 in APAC) | Wide-Mouth Round Carboy (2235-0050) | Narrow-Mouth Round Carboy with Tubulation 2301-0020) | Rectangular Carboy with Spigot (2321-0050) | Rectangular Carboy (2212-0050) |
Dispensing lab water and reagents. Compatible with sterilization by autoclaving | Liquid storage and shipping. Applications where autoclaving is required. | Liquid storage where autoclaving is required. Wide mouth allows easy access for filling and cleaning | For gravity fed liquid dispensing through tubing. Applications where autoclaving is required. | Dispensing lab water and reagents. Excellent choice for space efficient reagent banks. Autoclavable | Space-efficient storage for large-volume lab solutions where autoclaving is required |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PP with white PP closure/TPE gasket, PP spigot | PP with white PP closure/TPE gasket | PPCO with white PP closure | PP with white PP closure/TPE gasket | PPCO with white PP closure, PP spigot | PPCO with white PP closure |
Round | Round | Round | Round | Rectangular | Rectangular |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Wide mouth | Narrow mouth | Wide mouth | Wide mouth |
Spigot | No drain | No drain | Tubing adapter port | Spigot | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range 0°C to 135°C | Temp range 0°C to 135°C | Temp range -40°C to 121°C | Temp range 0°C to 135°C | Temp range -40°C to 121°C | Temp range -40°C to 121°C |
Autoclavable | Autoclavable | Autoclavable | Autoclavable | Autoclavable | Autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Stainless steel handle and 20L w/molded-in grip | Stainless steel handle and 20L w/molded-in grip |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
BPA-free | BPA-free | BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-mouth Round Carboy with Spigot (2319-0020, 8319-0020 APAC) | Narrow-Mouth Round Carboy (2250-0020, 8250-series in APAC) | Wide-Mouth Round Carboy (2235-0020) | Narrow-Mouth Round Carboy with Tubulation Port (2301-0020) | Rectangular Carboy with Spigot (2321-0020) | Rectangular Carboy (2212-0020) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g., over 100°C / 212°F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Rectangular Carboy with Spigot (2320-0050) | Rectangular Carboy (2211-0050) | Heavy-Duty Rectangular Carboy (2214-0050) | Rectangular Carboy with Tubulation (2303-0050) | Single Use Sterile Carboy (342289-series) |
Dispensing lab water and reagents. Excellent choice for space efficient reagent banks. | Space-efficient storage for large volume lab solutions. | Space-saving, extra-durable design for storing and shipping large volume lab reagents and other liquids. | Dispensing media, DI water and other lab reagents by gravity feed. Rectangular design optimizes shelf space | Supplied sterile for one-time use, the high-density polyethylene vessels are ideal for storing sterile fluids and pharmaceutical/biotech products. |
![]() | ![]() | ![]() | ![]() | ![]() |
HDPE with PP spigot and white PP closure | HDPE with white PP closure | HDPE with white PP closure | HDPE with white PP closure | HDPE with white PP closure |
Rectangular | Rectangular | Rectangular | Rectangular | Round |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga)and 33L (8.7ga) |
Wide mouth | Wide mouth | Narrow mouth | Wide mouth | Narrow mouth |
Spigot | No drain | No drain | Tubing adapter port | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile | Sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | No graduations |
Stainless steel handle | Stainless steel handle and 20L w/molded-in grip | Molded-in handle | Stainless steel handle and molded-in grip | Stainless steel handle sold separately/molded-in shoulder handle |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-free | BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Rectangular Carboy with Spigot (2320-0050) | Rectangular Carboy (2211-0050) | Heavy-Duty Rectangular Carboy (2214-0050) | Rectangular Carboy with Tubulation (2303-0050) | Single Use Sterile Carboy (342289-series) | |
Acids, dilute or weak | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E |
Aldehydes | G | G | G | G | G |
Bases/alkali | E | E | E | E | E |
Esters | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot (2318-series, 8318-series in APAC) | Narrow-Mouth Round Carboy (2210-series, 8210-series in APAC) | Narrow-Mouth Round Carboy with Tubulation Port (2302-0050) | Wide-Mouth Round Carboy (2234-0050) |
Classic DI water and lab reagent dispenser | Classic DI water and reagent reservoir | For gravity fed liquid dispensing through tubing | Wide mouth reagent reservoir provides easy access and cleaning |
![]() | ![]() | ![]() | ![]() |
LDPE carboy with white PP closure and PP spigot | LDPE carboy with white PP closure | LDPE carboy with white PP closure | LDPE carboy with white PP closure |
Round | Round | Round | Round |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5.5ga) and 25L (6.5ga) | 20L (5ga) and 25L (6.5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Narrow mouth | Wide mouth |
Spigot | No drain | Tubing adapter port | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -76°C to 70°C | Temp range -76°C to 70°C | Temp range -76°C to 70°C | Temp range -76°C to 70°C |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Round Carboy with Spigot (2318-series, 8318-series in APAC) | Narrow-Mouth Round Carboy (2210-series, 8210-series in APAC) | Narrow-Mouth Round Carboy with Tubulation Port (2302-0050) | Wide-Mouth Round Carboy (2234-0050) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Narrow-Mouth Round Clearboy with Spigot (2317-0050) | Narrow-Mouth Round Clearboy Carboy (2251-0050) | Wide-Mouth Rectangular Clearboy with Spigot (2322-0050) | Wide-Mouth Rectangular Clearboy (DS2213-0050) |
Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Compatible with sterilization. | Mixing, storing and shipping lab media and non-aggressive aqueous reagents where a clear view of contents.. | Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimized shelf space. | Storing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimizes shelf space. |
![]() | ![]() | ![]() | ![]() |
PC with white PP closure/TPE gasket and PP spigot | PC with white PP closure/TPE gasket | PC with white PP closure and PP spigot | PC with white PP closure |
Round | Round | Rectangular | Rectangular |
Clear | Clear | Clear | Clear |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Wide mouth | Wide mouth |
Spigot | No drain | Spigot | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -135°C to 135°C | Temp range -135°C to 135°C | Temp range -135°C to 135°C | Temp range -135°C to 135°C |
Autoclavable | Autoclavable | Autoclavable | Autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
No handles | No handles | Stainless steel handle | Stainless steel handle |
Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use |
Contains BPA | Contains BPA | Contains BPA | Contains BPA |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Round Clearboy with Spigot (2317-0050) | Narrow-Mouth Round Clearboy Carboy (2251-0050) | Wide-Mouth Rectangular Clearboy with Spigot (2322-0050) | Wide-Mouth Rectangular Clearboy (DS2213-0050) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | N | N | N | N |
Alcohols, aliphatic | G | G | G | G |
Aldehydes | F | F | F | F |
Bases/alkali | N | N | N | N |
Esters | N | N | N | N |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Fluorinated Carboy (2097-0050) | Fluorinated Carboy with Spigot (DS2327-0050) |
Storage for solvents and other organic solutions | Space-efficient dispensing for non-flammable solvents and other organic solutions |
![]() | ![]() |
FLPE with white PP closure | FLPE with white PP closure and fluoropolymer spigot |
Round | Rectangular |
Milky white translucent, yellowish tint due to fluorination process | Milky white translucent, yellowish tint due to fluorination process |
20L (5ga) | 20L (5ga) |
Narrow mouth | Wide mouth |
No drain | Spigot |
Leakproof* | Leakproof* |
Temp range -100°C to 120°C | Temp range -100°C to 120°C |
Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handle | Stainless steel handle |
Complies with Part 21 CFR 177.1615 for food use1 | Complies with Part 21 CFR 177.1615 for food use1 |
BPA-free | BPA-free |
Phthalate-free | Phthalate-free |
Fluorinated Carboy (2097-0050) | Fluorinated Carboy with Spigot (DS2327-0050) | |
Acids, dilute or weak | E | E |
Acids^ strong, concentrated | G | G |
Alcohols, aliphatic | E | E |
Aldehydes | G | G |
Bases/alkali | F | F |
Esters | G | G |
Hydrocarbons, aliphatic | E | E |
Hydrocarbons, aromatic | E | E |
Hydrocarbons, halogenated | G | G |
Ketones, aromatic | G | G |
Oxidizing agents, strong | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot (2319-0050, 8319-0050 APAC) | Narrow-Mouth Round Carboy (2250-0050, 8250-0050 in APAC) | Wide-Mouth Round Carboy (2235-0050) | Narrow-Mouth Round Carboy with Tubulation 2301-0020) | Rectangular Carboy with Spigot (2321-0050) | Rectangular Carboy (2212-0050) |
Dispensing lab water and reagents. Compatible with sterilization by autoclaving | Liquid storage and shipping. Applications where autoclaving is required. | Liquid storage where autoclaving is required. Wide mouth allows easy access for filling and cleaning | For gravity fed liquid dispensing through tubing. Applications where autoclaving is required. | Dispensing lab water and reagents. Excellent choice for space efficient reagent banks. Autoclavable | Space-efficient storage for large-volume lab solutions where autoclaving is required |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
PP with white PP closure/TPE gasket, PP spigot | PP with white PP closure/TPE gasket | PPCO with white PP closure | PP with white PP closure/TPE gasket | PPCO with white PP closure, PP spigot | PPCO with white PP closure |
Round | Round | Round | Round | Rectangular | Rectangular |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Wide mouth | Narrow mouth | Wide mouth | Wide mouth |
Spigot | No drain | No drain | Tubing adapter port | Spigot | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range 0°C to 135°C | Temp range 0°C to 135°C | Temp range -40°C to 121°C | Temp range 0°C to 135°C | Temp range -40°C to 121°C | Temp range -40°C to 121°C |
Autoclavable | Autoclavable | Autoclavable | Autoclavable | Autoclavable | Autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Stainless steel handle and 20L w/molded-in grip | Stainless steel handle and 20L w/molded-in grip |
Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use |
BPA-free | BPA-free | BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-mouth Round Carboy with Spigot (2319-0020, 8319-0020 APAC) | Narrow-Mouth Round Carboy (2250-0020, 8250-series in APAC) | Wide-Mouth Round Carboy (2235-0020) | Narrow-Mouth Round Carboy with Tubulation Port (2301-0020) | Rectangular Carboy with Spigot (2321-0020) | Rectangular Carboy (2212-0020) | |
Acids, dilute or weak | E | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E | E |
Aldehydes | G | G | G | G | G | G |
Bases/alkali | E | E | E | E | E | E |
Esters | G | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g., over 100°C / 212°F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Rectangular Carboy with Spigot (2320-0050) | Rectangular Carboy (2211-0050) | Heavy-Duty Rectangular Carboy (2214-0050) | Rectangular Carboy with Tubulation (2303-0050) | Single Use Sterile Carboy (342289-series) |
Dispensing lab water and reagents. Excellent choice for space efficient reagent banks. | Space-efficient storage for large volume lab solutions. | Space-saving, extra-durable design for storing and shipping large volume lab reagents and other liquids. | Dispensing media, DI water and other lab reagents by gravity feed. Rectangular design optimizes shelf space | Supplied sterile for one-time use, the high-density polyethylene vessels are ideal for storing sterile fluids and pharmaceutical/biotech products. |
![]() | ![]() | ![]() | ![]() | ![]() |
HDPE with PP spigot and white PP closure | HDPE with white PP closure | HDPE with white PP closure | HDPE with white PP closure | HDPE with white PP closure |
Rectangular | Rectangular | Rectangular | Rectangular | Round |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga)and 33L (8.7ga) |
Wide mouth | Wide mouth | Narrow mouth | Wide mouth | Narrow mouth |
Spigot | No drain | No drain | Tubing adapter port | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C | Temp range -100°C to 120°C |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile | Sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | No graduations |
Stainless steel handle | Stainless steel handle and 20L w/molded-in grip | Molded-in handle | Stainless steel handle and molded-in grip | Stainless steel handle sold separately/molded-in shoulder handle |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-free | BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Rectangular Carboy with Spigot (2320-0050) | Rectangular Carboy (2211-0050) | Heavy-Duty Rectangular Carboy (2214-0050) | Rectangular Carboy with Tubulation (2303-0050) | Single Use Sterile Carboy (342289-series) | |
Acids, dilute or weak | E | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G | G |
Alcohols, aliphatic | E | E | E | E | E |
Aldehydes | G | G | G | G | G |
Bases/alkali | E | E | E | E | E |
Esters | G | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N | N |
Ketones, aromatic | N | N | N | N | N |
Oxidizing agents, strong | F | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Narrow-Mouth Round Carboy with Spigot (2318-series, 8318-series in APAC) | Narrow-Mouth Round Carboy (2210-series, 8210-series in APAC) | Narrow-Mouth Round Carboy with Tubulation Port (2302-0050) | Wide-Mouth Round Carboy (2234-0050) |
Classic DI water and lab reagent dispenser | Classic DI water and reagent reservoir | For gravity fed liquid dispensing through tubing | Wide mouth reagent reservoir provides easy access and cleaning |
![]() | ![]() | ![]() | ![]() |
LDPE carboy with white PP closure and PP spigot | LDPE carboy with white PP closure | LDPE carboy with white PP closure | LDPE carboy with white PP closure |
Round | Round | Round | Round |
Milky white translucent | Milky white translucent | Milky white translucent | Milky white translucent |
20L (5.5ga) and 25L (6.5ga) | 20L (5ga) and 25L (6.5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Narrow mouth | Wide mouth |
Spigot | No drain | Tubing adapter port | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -76°C to 70°C | Temp range -76°C to 70°C | Temp range -76°C to 70°C | Temp range -76°C to 70°C |
Not autoclavable | Not autoclavable | Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles | Molded-in shoulder handles |
Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 |
BPA-free | BPA-free | BPA-free | BPA-free |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Round Carboy with Spigot (2318-series, 8318-series in APAC) | Narrow-Mouth Round Carboy (2210-series, 8210-series in APAC) | Narrow-Mouth Round Carboy with Tubulation Port (2302-0050) | Wide-Mouth Round Carboy (2234-0050) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | G | G | G | G |
Alcohols, aliphatic | E | E | E | E |
Aldehydes | G | G | G | G |
Bases/alkali | E | E | E | E |
Esters | G | G | G | G |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Narrow-Mouth Round Clearboy with Spigot (2317-0050) | Narrow-Mouth Round Clearboy Carboy (2251-0050) | Wide-Mouth Rectangular Clearboy with Spigot (2322-0050) | Wide-Mouth Rectangular Clearboy (DS2213-0050) |
Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Compatible with sterilization. | Mixing, storing and shipping lab media and non-aggressive aqueous reagents where a clear view of contents.. | Dispensing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimized shelf space. | Storing lab media and non-aggressive aqueous lab reagents where a clear view of contents is desired. Rectangular shape optimizes shelf space. |
![]() | ![]() | ![]() | ![]() |
PC with white PP closure/TPE gasket and PP spigot | PC with white PP closure/TPE gasket | PC with white PP closure and PP spigot | PC with white PP closure |
Round | Round | Rectangular | Rectangular |
Clear | Clear | Clear | Clear |
20L (5ga) | 20L (5ga) | 20L (5ga) | 20L (5ga) |
Narrow mouth | Narrow mouth | Wide mouth | Wide mouth |
Spigot | No drain | Spigot | No drain |
Leakproof* | Leakproof* | Leakproof* | Leakproof* |
Temp range -135°C to 135°C | Temp range -135°C to 135°C | Temp range -135°C to 135°C | Temp range -135°C to 135°C |
Autoclavable | Autoclavable | Autoclavable | Autoclavable |
Non-sterile | Non-sterile | Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons | Graduated in liters and gallons |
No handles | No handles | Stainless steel handle | Stainless steel handle |
Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use | Complies with Part 21 CFR 177.1580 for food use |
Contains BPA | Contains BPA | Contains BPA | Contains BPA |
Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
Narrow-Mouth Round Clearboy with Spigot (2317-0050) | Narrow-Mouth Round Clearboy Carboy (2251-0050) | Wide-Mouth Rectangular Clearboy with Spigot (2322-0050) | Wide-Mouth Rectangular Clearboy (DS2213-0050) | |
Acids, dilute or weak | E | E | E | E |
Acids^ strong, concentrated | N | N | N | N |
Alcohols, aliphatic | G | G | G | G |
Aldehydes | F | F | F | F |
Bases/alkali | N | N | N | N |
Esters | N | N | N | N |
Hydrocarbons, aliphatic | G | G | G | G |
Hydrocarbons, aromatic | N | N | N | N |
Hydrocarbons, halogenated | N | N | N | N |
Ketones, aromatic | N | N | N | N |
Oxidizing agents, strong | F | F | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
Fluorinated Carboy (2097-0050) | Fluorinated Carboy with Spigot (DS2327-0050) |
Storage for solvents and other organic solutions | Space-efficient dispensing for non-flammable solvents and other organic solutions |
![]() | ![]() |
FLPE with white PP closure | FLPE with white PP closure and fluoropolymer spigot |
Round | Rectangular |
Milky white translucent, yellowish tint due to fluorination process | Milky white translucent, yellowish tint due to fluorination process |
20L (5ga) | 20L (5ga) |
Narrow mouth | Wide mouth |
No drain | Spigot |
Leakproof* | Leakproof* |
Temp range -100°C to 120°C | Temp range -100°C to 120°C |
Not autoclavable | Not autoclavable |
Non-sterile | Non-sterile |
Graduated in liters and gallons | Graduated in liters and gallons |
Molded-in shoulder handle | Stainless steel handle |
Complies with Part 21 CFR 177.1615 for food use1 | Complies with Part 21 CFR 177.1615 for food use1 |
BPA-free | BPA-free |
Phthalate-free | Phthalate-free |
Fluorinated Carboy (2097-0050) | Fluorinated Carboy with Spigot (DS2327-0050) | |
Acids, dilute or weak | E | E |
Acids^ strong, concentrated | G | G |
Alcohols, aliphatic | E | E |
Aldehydes | G | G |
Bases/alkali | F | F |
Esters | G | G |
Hydrocarbons, aliphatic | E | E |
Hydrocarbons, aromatic | E | E |
Hydrocarbons, halogenated | G | G |
Ketones, aromatic | G | G |
Oxidizing agents, strong | F | F |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
E | No damage after 30 days constant exposure at 20oC. |
G | Little or no damage after 30 days constant exposure at 20oC. |
F | Some effect after 7 days constant exposure at 20oC. |
N | Not recommended. Immediate damage may occur. |
Footnote:
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see "Oxidizing acids, strong."
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to
techsupport.labproducts.eu@thermofisher.com.
Regulatory Support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at RocRegSupport@thermofisher.com
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.