pHrodo pH sensors for detecting cytosolic and vesicle pH
(See a list of the products featured in this article.)
Intracellular pH is one of a cell’s essential control switches for regulating critical cell functions. Not only does pH affect enzyme activity and other protein functions, but proton gradients are a critical source of energy for driving cell metabolism. Eukaryotic cells contain a variety of defined compartments with different degrees of acidity: intracellular pH is generally between ~6.8 and 7.2 in the cytosol and between ~4.5 and 6.5 in acidic vesicles and organelles. The pH-sensitive pHrodo dyes are unique pH sensors that undergo a dramatic increase in fluorescence in response to an environmental shift from high to low pH. With a pKa of approximately 6.8, both the pHrodo Red and pHrodo Green dyes are useful pH indicators between pH 4 and pH 8, making them ideal for monitoring physiologically relevant pH changes.
The pHrodo Red and pHrodo Green dyes are only dimly fluorescent above pH 8; however, as the pH of their surroundings becomes more acidic, their fluorescence intensity increases significantly [1] (Figure 1). As a consequence, pHrodo Red fluorescence (excitation/emission maxima ~566/590 nm) and pHrodo Green fluorescence (excitation/emission maxima ~505/520 nm) provide a positive indication of processes such as apoptosis, phagocytic ingestion, and lysosomal sequestration that require acidification, in contrast to the reduction in fluorescence exhibited by fluorescein derivatives. Furthermore, with both red- and green-fluorescent pHrodo dyes available, you have more flexibility in the design of multiplex cellular assays. Both pHrodo dyes can be excited with 488 nm light and are compatible with argon-ion lasers commonly installed in flow cytometers, microscopes, and microplate readers.
pH response profile response graph of pHrodo® Red and pHrodo® Green dextrans 
Figure 1. pH response profile of pHrodo Red and pHrodo Green dextrans.
pHrodo Red dextran and pHrodo Green dextran were monitored at excitation/emission wavelengths of 550/585 nm and 505/525 nm, respectively, in a fluorescence microplate reader. Citrate, MOPS, and borate buffers were used to span the pH range from 4 to 8.5.
Detect and calibrate cytosolic pH with pHrodo AM dyes
To enable cytosolic pH measurements, we have developed cell-permeant forms of the pHrodo Red and pHrodo Green dyes (Figure 2). The pHrodo Red AM and pHrodo Green AM intracellular pH indicators have been modified with acetoxymethyl (AM) ester groups to produce an uncharged molecule that can permeate cell membranes. Once inside the cell, these lipophilic blocking groups are cleaved by nonspecific esterases, resulting in a pH-sensitive dye that is retained within the intracellular space. Unlike the traditional fluorescein-based pH indicator BCECF AM, pHrodo Red AM and pHrodo Green AM exhibit increasing fluorescence as pH decreases and do not photobleach significantly (Figure 3).
To calibrate the fluorescence exhibited by cells loaded with pHrodo AM dyes (or other intracellular pH indicators), we offer the Intracellular pH Calibration Buffer. This kit contains a selection of calibrated pH solutions (pH 4.5, 5.5, 6.5, and 7.5), as well as the ionophores nigericin and valinomycin, which help equilibrate the pH inside and outside of the cells. After using pHrodo Red AM or pHrodo Green AM—or any of the pHrodo conjugates—and measuring fluorescence intensity under experimental conditions, the cells are resuspended in one of the calibration buffers to which valinomycin and nigericin have been added, and the fluorescence intensity is measured again. This process is repeated with the remaining calibration buffers so that a standard curve can be created and used to pinpoint the intracellular pH associated with the experimental conditions (Figure 4).
Two panel figure of intracellular pH visualized in live U2OS cells incubated with pHrodo® Red AM and pHrodo® Green AM intracellular pH indicators Figure 2. Intracellular pH visualized in live cells. U2OS cells were incubated with (A) 5 μM pHrodo Red AM or (B) 10 μM pHrodo Green AM intracellular pH indicators using PowerLoad™ Concentrate (provided with the AM esters) for 30 min at room temperature, and then 30 min at 37°C. Both dyes provide an indication of relative pH in different cellular compartments. Acidic compartments and organelles fluoresce more brightly.
Two panel figure illustrating real-time intracellular pH measurements of NIH/3T3 cells loaded with BCECF AM and pHrodo® Red AM intracellular pH indicators 
Figure 3. Real-time intracellular pH measurements. NIH/3T3 cells were loaded with (A) 5 μM BCECF AM or (B) 5 μM pHrodo Red AM intracellular pH indicators. Cells were then washed with a series of HEPES-based pH standard buffers containing 10 μM nigericin and 10 μM valinomycin to clamp intracellular pH to the indicated values. BCECF AM fluorescence decreases stepwise as pH is reduced, and decreases continuously due to photobleaching. In contrast, pHrodo Red AM fluorescence increases as pH is reduced. Furthermore, pHrodo Red AM does not photobleach significantly after 12 min of imaging.
Two panel figure of standard curves showing the linear relationship between intracellular pH and relative fluorescence in U2OS cells incubated with pHrodo® Red and pHrodo® Green AM intracellular pH indicators 
Figure 4. Intracellular pH calibration. U2OS cells in 96-well plates were incubated with (A) 5 μM pHrodo Red AM or (B) 10 μM pHrodo Green AM intracellular pH indicators. Standard buffers from the Intracellular pH Calibration Buffer Kit were used to clamp the intracellular pH to 4.5, 5.5, 6.5, and 7.5. High-content screening was used to measure mean cellular fluorescence of triplicate samples. A standard curve shows the linear relationship between intracellular pH and relative fluorescence. 
Visualizing intracellular pH during apoptosis
A distinctive feature of the early stages of apoptosis is the acidification of the cytosol, followed by caspase activation and the eventual breakdown of plasma membrane integrity [2, 3]. We have used the intracellular pH indicator pHrodo Red AM in conjunction with the CellEvent Caspase-3/7 Green Detection Reagent (a fluorogenic caspase substrate) to monitor a population of HeLa cells undergoing apoptosis. The time course in Figure 5 shows a single cell over the course of 4 hours after treatment with camptothecin. This cell first exhibits increasing pHrodo Red fluorescence, indicating the acidification of the cytosol, followed by the appearance of the green fluorescence from the caspase-3/7 probe. An adjacent cell was apparently not induced by the camptothecin treatment and remains only dimly fluorescent throughout the time course.
10-panel figure showing temporal resolution of intracellular acidification and caspase-3/7 activation in a HeLa cell undergoing apoptosis over time 
Figure 5. Temporal resolution of intracellular acidification and caspase-3/7 activation in a HeLa cell undergoing apoptosis. HeLa cells were loaded with the intracellular pH indicator pHrodo Red AM (5 μM) and the fluorogenic caspase substrate CellEvent Caspase-3/7 Green Detection Reagent (2 μM) and then treated with 10 μM camptothecin to induce apoptosis. Cells were imaged continuously over 4 hr on the EVOS FL Auto Imaging System using a 40x objective and the EVOS Onstage Incubator. The cell undergoing apoptosis shows increasing pHrodo Red fluorescence as the intracellular pH drops in the earliest stages of apoptosis, followed by increasing green fluorescence as caspase-3 and -7 are activated. The apparent loss of the pHrodo Red signal was likely due to compromised membrane integrity and subsequent loss of the dye from the cell. 
Detect and monitor pH changes during cellular internalization
The pHrodo Red and pHrodo Green dyes are also available as protein and dextran conjugates for studying cellular internalization mechanisms. The ability to study these internalization processes has historically been limited by the lack of tools to directly monitor the ingestion and subsequent acidification of extracellular matter by live cells. We have developed pHrodo Red and pHrodo Green dye conjugates that are designed for studying the three primary mechanisms of endocytosis in live cells: 1) pHrodo conjugates of EGF and transferrin can be used to monitor receptor-mediated endocytosis, 2) pHrodo dextran conjugates can be used to study fluid-phase endocytosis, and 3) pHrodo conjugates of bacteria and yeast can be used to track phagocytosis (Figure 6).
Alternatively, you can create your own pH-sensitive bioconjugates using amine- and thiol-reactive derivatives of the pHrodo Red and pHrodo Green dyes. Potential applications for pHrodo bioconjugates include tracking antibody internalization and monitoring virus infection after labeling viral coat proteins.
Flow cytometry analysis histogram showing increased fluorescence of granulocytes treated with pHrodo® Red BioParticles® conjugates Figure 6. Flow cytometry analysis showing increased fluorescence of granulocytes treated with pHrodo Red BioParticles conjugates. A whole blood sample was collected and treated with heparin, and two 100 μL aliquots were prepared. Both aliquots were treated with pHrodo Red E. coli BioParticles conjugates and vortexed. One sample was placed in a 37°C water bath, and the other sample (negative control) was placed in an ice bath. After a 15 min incubation, red blood cells were lysed with an ammonium chloride–based lysis buffer. The samples were centrifuged for 5 min at 500 x g, washed once, and resuspended in HBSS. The samples were then analyzed on a BD FACSCalibur™ cytometer using a 488 nm argon laser and 564–606 nm emission filter. The sample incubated at 37°C shows the increased fluorescence of the phagocytosed pHrodo Red BioParticles (red), in contrast to the negative control sample, which was kept on ice to inhibit phagocytosis (blue).
Explore the role of pH in cell health
pHrodo pH indicators are ideal live-cell probes for the study of intracellular pH and cellular internalization, and their regulation in both normal development and disease processes. With both red- and green-fluorescent pHrodo derivatives, you can more easily incorporate pHrodo dyes into multiplex experiments with other fluorescent live-cell probes, including MitoTracker mitochondrial probes, CellROX oxidative stress probes, fluorescent membrane markers, live-cell DNA stains, and CellLight targeted fluorescent proteins.
Ordering information
Catalog # Name Size Price (USD) Qty
P35373 pHrodo™ Green AM Intracellular pH Indicator, 50 μL Each
595.65
Online Exclusive
614.00
Save 18.35 (3%)
P35372 pHrodo™ Red AM Intracellular pH Indicator, 50 μL Each
598.65
Online Exclusive
620.00
Save 21.35 (3%)
P35380 pHrodo™ AM Variety Pack Each
284.00
P35379 Intracellular pH Calibration Buffer Kit Each
313.65
Online Exclusive
327.00
Save 13.35 (4%)
P35366 pHrodo™ Green E. coli BioParticles™ Conjugate for Phagocytosis Each
534.65
Online Exclusive
550.00
Save 15.35 (3%)
P35381 pHrodo™ Green E. coli BioParticles™ Phagocytosis Kit for Flow Cytometry Each
1,356.00
P35367 pHrodo™ Green S. aureus BioParticles™ Conjugate for Phagocytosis, 5 x 2 mg Each
527.65
Online Exclusive
552.00
Save 24.35 (4%)
P35382 pHrodo™ Green S. aureus BioParticles™ Phagocytosis Kit for Flow Cytometry Each
1,366.00
P35365 pHrodo™ Green Zymosan BioParticles™ Conjugate for Phagocytosis, 5 x 1 mg Each
532.65
Online Exclusive
552.00
Save 19.35 (4%)
P35364 pHrodo™ Red Zymosan BioParticles™ Conjugate for Phagocytosis Each
526.65
Online Exclusive
552.00
Save 25.35 (5%)
P35361 pHrodo™ Red E. coli BioParticles™ Conjugate for Phagocytosis, 5 x 2 mg Each
540.65
Online Exclusive
560.00
Save 19.35 (3%)
A10025 pHrodo™ Red E. coli BioParticles™ Phagocytosis Kit for Flow Cytometry, 100 assay kit Each
1,380.00
A10010 pHrodo™ Red S. aureus BioParticles™ Conjugate for Phagocytosis, 5 x 2 mg Each
534.65
Online Exclusive
554.00
Save 19.35 (3%)
A10026 pHrodo™ Red Phagocytosis Particle Labeling Kit for Flow Cytometry Each
1,382.00
P35368 pHrodo™ Green Dextran, 10,000 MW, for Endocytosis, 0.5 mg Each
536.65
Online Exclusive
556.00
Save 19.35 (3%)
P10361 pHrodo™ Red Dextran, 10,000 MW, for Endocytosis, 0.5 mg Each
553.65
Online Exclusive
574.00
Save 20.35 (4%)
P35375 pHrodo™ Green Epidermal Growth Factor (EGF) Conjugate, 20 μg Each
548.65
Online Exclusive
576.00
Save 27.35 (5%)
P35374 pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate, 20 μg Each
550.65
Online Exclusive
578.00
Save 27.35 (5%)
P35376 pHrodo™ Red Transferrin Conjugate, 1 mg Each
611.65
Online Exclusive
642.00
Save 30.35 (5%)
P35362 pHrodo™ Red Avidin (Fluorogenic pH Sensor) Each
682.00
P35370 pHrodo™ Green Maleimide Each
780.00
P35369 pHrodo™ Green STP Ester Each
614.00
P35371 pHrodo™ Red Maleimide Each
790.00
P36600 pHrodo™ Red, succinimidyl ester (pHrodo™ Red, SE) Each
846.00
Article download
Download a hyperlink-enabled, printer-friendly version of this article.
Download now
For Research Use Only. Not for use in diagnostic procedures.