Spin with confidence
Reliable and efficient centrifuges
Thermo Scientific clinical centrifuges are designed to deliver exceptional performance and precision, ensuring optimal workflow efficiency for a wide range of common clinical applications. Our benchtop centrifuges provide a compact footprint with capacities from 120 mL to 4 L and a comprehensive rotor portfolio from 1.5 mL to 50 mL sample tubes, microplates, and 10 mL standard syringes. With innovative technologies and a flexible design, they help simplify the clinical sample preparation process.
Medifuge Centrifuge
Designed for routine clinical and life science applications, the Thermo Scientific Medifuge Small Bench Centrifuge offers a compact footprint and features the unique Thermo Scientific DualSpin rotor with its hybrid design. This allows to quickly change between fixed angle and swinging buckets using the same rotor.
Medifuge Brochure
Fits in and stands out
Medifuge Application Note
Coagulation Testing through preparation of Platelet-Poor Plasma (PPP)
Medifuge Green Fact Sheet
Generates up to 62% less waste at end of life.
Other Benchtop Centrifuges for Clinical Applications
Microcentrifuges
Small Bench Centrifuges
Compact General Purpose Centrifuges
High-Capacity General Purpose Centrifuges
Description
A sleek, remarkably small footprint provides outstanding processing power, with a rugged design for years of dependability.
  • Process up to 24 microtubes, pediatric blood tubes or PCR tubes per run
  • ClickSeal biocontainment lids provide added safety
Compact centrifuges for effective and efficient processing of clinical samples, including platelet-poor plasma tests.
  • Process up to 24 x 5/7mL blood tubes
  • ClickSeal biocontainment lids provide added safety
  • Run platelet-poor plasma tests in 1 minute with the Thermo Scientific HIGHConic rotor
For clinical laboratories requiring mid-level throughput in a small footprint, these 1.6L compact benchtop centrifuges are a great solution. Outfitted with Auto-Lock to quickly and easily remove rotors to maximize versatility and facilitate convenient cleaning.
  • Process up to 84 x 5/7 mL blood tubes
  • ClickSeal biocontainment lids provide added safety
  • 15 rotor options available for most common clinical applications
Achieve 4L capacity in a 3L footprint in this high-capacity centrifuge, available in benchtop or floor-standing options. Outfitted with Auto-Lock to quickly and easily remove rotors to maximize versatility and facilitate convenient cleaning.
  • Process up to 196 x 5/7mL blood tubes
  • ClickSeal biocontainment lids provide added safety
  • 13 rotor options available for most common clinical applications
Models
Floor standing model:
Floor standing models:
Application Overview
  Clinical
Chemistry/
Hematology
Pathology Clinical
Microbiology
Molecular
Biology
Hematology Clinical
Immunology
Pediatrics
Thermo Scientific
Centrifuges
RBC/
Coagulation
Studies
Urinalysis
and
Platelet
Poor
Plasma
TB
Sputum
Chlamydia
Trachomatis
DNA/
RNA
Isolation
Hematocrit Leucocytes ELISA/
RIA
Blood
Analysis
Medifuge centrifuge            
Microcentrifuges              
Small bench centrifuges
Compact general purpose centrifuges  
High-capacity general purpose centrifuges  
Blood Banking Centrifuges
Thermo Scientific Large Capacity Centrifuges provide medium to high throughput processing with a variety of swinging bucket rotors, so you can use your validated protocols to process a range of products in blood bags or tubes. Our products are certified in accordance with the EU 2017/745 Medical Device Regulation.
Large capacity centrifuges
For regular- to medium-throughput blood processing needs, the Sorvall BP 8 / Cryofuge 8 centrifuge provides 6 x 550mL or 8 x 550mL blood bag capacity, utilizing the same bucket system. The Sorvall BP 16 / Cryofuge 16 centrifuge is designed for high-throughput blood processing with its 12 x 550mL capacity and is upgradeable to 16 x 550mL blood bags with the same bucket selections. Our large capacity centrifuges offer flexibility to meet your needs with a range of buckets and adapters to spin several types of tubes. 
Order now
Resources
Video
Clinical Centrifuge Solutions
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00