Understanding the intricacies of mitochondrial function is crucial for providing valuable insights in cellular processes such as programmed cell death. Thermo Fisher Scientific offers a comprehensive range of reagents and assays that detect and monitor changes in mitochondria function for imaging and flow cytometry platforms. Our products are suitable for real-time measurements in live cells as well as compatible with fixation and immunodetection. They enable researchers to assess key parameters such as such as mitochondrial membrane potential, superoxide production, calcium levels, and mitochondrial permeability transition pore activity.
Mitochondrial function changes in apoptosis
A feature of the early stages of programmed cell death is the disruption of mitochondrial function. This mitochondrial disruption includes changes in the membrane potential - a central feature of mitochondrial health, and alterations to the oxidation–reduction potential of the mitochondria. The inner mitochondrial membrane potential is essential in Ca2+ uptake and storage, reactive oxygen species generation, detoxification, and most importantly, the synthesis of ATP by oxidative phosphorylation (1). Therefore, the membrane’s depolarization is a good indicator of mitochondrial dysfunction, which is increasingly implicated in drug toxicity (2-6). Changes in the membrane potential, along with decreases ATP to ADP ratios, increased mitochondrial matrix calcium levels, oxidative stress, and release of cytochrome c into the cytosol, are all presumed to be associated with the mitochondrial permeability transition, resulting in disruption of ions and small molecule homeostasis via the mitochondrial permeability transition pore (MPTP).
Mitochondria function assays
The disruption of mitochondria function can be detected using a variety of fluorescence-based assays including measurements of mitochondrial calcium, superoxide, mitochondrial permeability transition, and membrane potential. A summary of these assays are listed in Table 1 below.
Although disruption of mitochondrial function is associated with apoptosis, the various aspects listed above are not specific to apoptotic cell death. Because the aforementioned apoptosis-related mitochondria function parameters are not specific for apoptosis and may vary across cell types, it is often advantageous to consider multi-parametric assays, including the use of specific markers for the activation of particular caspase proteases.
Summary of mitochondria function assays
 Mitochondria function assays
 Membrane potential and MetabolismSuperoxide productionCalciumMitochondrial permeability transition
What can be identified?
Cells with healthy, metabolically active mitochondria.
2 options available:
Detection of dynamic changes in relative mitochondria membrane potential (reversible detection).
End point assays for detecting relative mitochondria membrane potential at a specific timepoint (irreversible detection).
Irreversible detection of mitochondrial superoxide production in live cells.
Irreversible detection of mitochondrial calcium in live cells.Irreversible detection of the mitochondrial permeability transition (MPT) subsequent to dysregulation of the MPT pore.
What is the basis of assay?Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.Measure the increasing fluorescence of a superoxide detection reagent that accumulates in mitochondria in a membrane potential-dependent fashion where it is selectively oxidized and subsequently binds nucleic acids.Measure the increasing fluorescence of a calcium indicator with increasing calcium concentration in the mitochondria.Loss of mitochondrial calcein fluorescence due to mitochondrial permeability transition.
Is the assay fixable?
Only a small selection of the end point assays for detecting mitochondria membrane potential are fixable. Please see the table of MitoTracker probes for Imaging and Flow Cytometry below.
None of the dynamic assays are fixable.
Not compatible with fixation.
Not compatible with fixation.
Not compatible with fixation.
Detection of dynamic changes in mitochondrial membrane potential
What can be identified?
Cells with healthy, metabolically active mitochondria. Dynamic changes detect relative mitochondria membrane potential (reversible detection). None of the dynamic assays are fixable.
What is the basis of the assay?
Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.
Dual-emission, ratiometric reagents
 JC-1MitoProbe JC-1 Assay KitMitoProbe DiOC2(3) Assay Kit
ReadoutActive mitochondria exhibit brighter red fluorescence signal compared to mitochondria with lower membrane potential which fluoresce green. 
Ex/Em (nm)514/529 (monomer, green)485/497 (monomer, green)
514/590 (aggregate, red)485/650 (aggregate, far–red)
Suggested filtersTRITCFITC and PEFITC and APC
Instrument platformImaging microscopyFlow cytometryFlow cytometry
Sample typeLive cellsLive cellsLive cells
Compatibility with fixationNoNoNo
Format5 mgKit contents:
JC-1, DMSO
CCCP (a mitochondria membrane potential disrupter in DMSO)
10x PBS
Kit contents:
DiOC2(3) in DMSO
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Cat. No.T3168M34152M34150
Single-emission reagents
 TMRMImage-iT TMRMMitoProbe TMRM Assay KitMitoProbe DiIC1(5) Assay Kit
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)548/574638/658
Suggested filterTRITC~585/16 nmAlexa Fluor 647/APC
Instrument platformFluorescence microscopyFlow cytometry
Sample typeLive cellsLive cellsLive cellsLive cells
Compatibility with fixationNoNoNoNo
Format25 mg5 x 100 μLKit contents:
TMRM
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Kit contents:
DiIC1(5) in DMSO
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Cat. No.T668I34361M20036M34151
JC-1 Dye and the MitoProbe JC-1 Assay Kit
JC-1 is a cell-permeant dye widely used for detecting mitochondrial membrane potential in flow cytometry and microscopy platforms (Figure 1). JC-1 exhibits potential-dependent accumulate within the mitochondria, forming J-aggregates as the dye increases. This increase in potential will result in a shift from the green fluorescence (~529 nm) monomer to red fluorescence (~590 nm) J-aggregates. Alternatively, depolarization of the mitochondria is denoted by a decrease in the red/green fluorescence intensity ratio. 
Learn more about the JC-1 Dye for mitochondria membrane potential
A
Microscopic view of mitochondria stained with green-red fluorescence at 4 different time course points.
B
Dot plot of JC-1 stained cells showing 2 cell populations-healthy cells colored red and apoptotic cells colored green.
Figure 1. Detection of mitochondria membrane potential using JC-1 Dye.(A) NIH 3T3 fibroblasts stained with JC-1 Dye show the progressive loss of red J-aggregate fluorescence and cytoplasmic diffusion of green monomer fluorescence following exposure to hydrogen peroxide. Images show the same field of cells viewed before H2O2 treatment and 5, 10 and 20 min after treatment. The images were contributed by Ildo Nicoletti, Perugia University Medical School. (B)MitoProbe JC-1 Assay Kit was used to stain Jurkat cells (T-cell leukemia, human) which were then analyzed on a flow cytometer using 488 nm excitation with 530 nm and 585 nm bandpass emission filters. Green = apoptotic cells (reduced mitochondria membrane potential), red = normal cells.
MitoProbe DiOC2(3) Assay Kit
Another dye that can be used as a ratiometric probe for mitochondria membrane potential is DiOC2(3). The mechanism is similar to that of JC-1 Dye in that the monomer exhibits green fluorescence at lower concentrations where mitochondria membrane potential is low and the aggregate of the dye in more active mitochondria causes the emission to shift toward the red. In the case of DiOC2(3), the shift is farther in the red than JC-1, above 650 nm. The MitoProbe DiOC2(3) Assay Kit has been specifically developed to work in flow cytometry applications and includes a mitochondrial membrane-potential disrupter, CCCP.
TOP
TMRM and MitoProbe TMRM Assay Kit
Tetramethylrhodamine, methyl ester (TMRM) or the related TMRE (tetramethylrhodamine, ethyl ester) are small, cell-permeant dyes that accumulate in active mitochondria. If the cells are healthy and have functioning mitochondria, the signal is bright. Upon loss of the mitochondrial membrane potential, TMRM and TMRE accumulation ceases and the signal dims or is lost from the mitochondria. This ability to dynamically monitor changes in mitochondria membrane potential is a distinct advantage of this probe. TMRM and TMRE signals can be detected in live cells or isolate mitochondria with traditional and high content fluorescence microscopy, microplates, or by flow cytometry (Figure 2).
TMRM is available in different formats to fit your apoptosis assay needs. See the Selection Guide (Tab 2. Single-emission reagents) for detection of dynamic changes in mitochondria membrane potential using single-emission reagents. MitoProbe TMRM Assay Kit for Flow Cytometry includes TMRM along with CCCP, a mitochondrial membrane potential disrupter and detailed protocol for use in flow cytometry.
A
Microscopic view of cells stained with orange fluorescence (mitochondria) and green fluorescence (tubulin)
B
Dot plot showing healthy, apoptotic and dead cells with and without treatment.
Figure 2. TMRM as a mitochondrial membrane potential assay for apoptosis.(A) HeLa cells labeled using Tubulin Tracker Green and Image-iT TMRM Reagent show superb multiplexing capability and staining specificity. Image shows multiple live mitotic cells with microtubules assembled into a mitotic spindle with visible microtubule filaments, as well as cleavage furrow indicating completion of cytokinesis. (B) Jurkat cells, a human T-lymphocyte cell line, were treated with DMSO (control) or 500 nM staurosporine for 2 hours. Cells were subsequently stained with MitoProbe TMRM for 30 min at 37˚C, followed by a wash and additional stain with Annexin V Pacific Blue conjugate. Staurosporine induced apoptosis, resulting in a mixed population of cells containing a population of healthy MitoProbe TMRM-positive cells as well as a population of apoptotic, Annexin V Pacific Blue-positive, MitoProbe TMRM-low cells.
TOP
MitoProbe DiIC1(5) dye
Cationic carbocyanine dyes have been shown to accumulate in cells in response to membrane potential. The MitoProbe DiIC1(5) Kit is designed for flow cytometry applications and provides the far-red–fluorescent DiIC1(5) carbocyanine dye, along with a mitochondrial membrane potential disrupter, CCCP. At concentrations below 100 nm, the cationic DiIC1(5) dye passively accumulates in the mitochondria with active membrane potentials in living cells (Figure 3). Unlike DiOC2(3), however, DiIC1(5) dye exhibits a single wavelength emission whose fluorescence signal increases with more active mitochondria.
Flow cytometry histogram showing cells with active mitochondria in control compared to cells with depolarized mitochondria with CCCP treatment
Figure 3. Detection of changes in mitochondrial membrane potential using the MitoProbe DiIC1(5) Assay Kit for Flow Cytometry. Decrease in mitochondrial membrane potential as demonstrated with DiIC1(5) fluorescence due to the addition of carbonyl cyanide 3-chlorophenylhydrazone (CCCP). Jurkat cells (T-cell leukemia, human) were stained with 50 nM DiIC1(5) alone (blue) or in the presence of 50 μM CCCP (red).
TOP
End point assays in mitochondrial membrane potential
What can be identified?
Cells with healthy, metabolically active mitochondria. End point assays detect relative mitochondria membrane potential at a specific timepoint (irreversible detection).
Only a small selection of the end point assays for detecting mitochondria membrane potential are fixable.
What is the basis of the assay?
Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.
 MitoTracker Probes—Imaging
MitoTracker Green FMMitoTracker Orange CMTMRosMitoTracker Red CMXRosMitoTracker Red FMMitoTracker Deep Red FM
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)490/516550/580579/599581/644644/665
Suggested filterFITCTRITCTexas RedCy5Cy5
Instrument platformFluorescence microscopy
Microplate reader
Sample typeLive cellsLive cellsLive cellsLive cellsLive cells
Compatibility with fixation NoYesYesNoNo
Format20 x 50 μg20 x 50 μg20 x 50 μg20 x 50 μg20 x 50 μg
See all MitoTracker dyes
 MitoTracker Probes—Flow Cytometry
MitoTracker Green FMMitoTracker Red CMXRosMitoTracker Red FMMitoTracker Deep Red FM
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)490/516579/599581/644644/665
Suggested filterFITCTexas RedCy5Cy5
Instrument platformFlow cytometry
Sample typeLive cellsLive cellsLive cellsLive cells
Compatibility with fixation*NoYesNoNo
Format5 x 20 μg5 x 20 μg5 x 20 μg5 x 20 μg
Cat. No.M7514M46752M46751M46753
MitoTracker probes
MitoTracker probes are small (<1 kDa), cell-permeant mitochondrion-selective dyes that contain a thiol-reactive chloromethyl moiety that keeps the dye associated with the mitochondria after fixation. Because the probes form covalent bonds with the mitochondrial thiols, they can only be used as an end point assays to detect mitochondrial membrane potential at the time of loading in live cells, and should not be used as sensors of dynamic mitochondrial membrane changes over time.
Impact of fixation on MitoTracker probes
While the MitoTracker signal can be fixed after labeling, in assays to study mitochondrial structure, if studying the mitochondrial membrane function, it is strongly suggested to use a few, select MitoProbe reagents. Only MitoTracker Orange CMTMRos and MitoTracker Red CMXRos have been found to maintain a consistent difference in signal loss when comparing mitochondrial membrane potential in control and treated samples after fixation. These single wavelength emission reagents can be used for flow cytometry, imaging microscopy, and high content analysis applications (Figures 4 and 5).
Learn more about MitoTracker probes for mitochondria structure
Flow cytometry dot plot showing separation of apoptotic and healthy cells in treated versus untreated cells.
 Click image to enlarge
Figure 4. MitoTracker Red CMXRos dye mitochondria membrane potential detection. Jurkat human T-cell leukemia cells in complete medium were (A) first exposed to 10 µM camptothecin for 4 hours or (B) left untreated. Both cell populations were then treated with the reagents in the Mitochondrial Membrane Potential/Annexin V Apoptosis Kit and analyzed by flow cytometry. Note that the apoptotic cells show higher reactivity for annexin V and lower MitoTracker Red dye fluorescence than do live cells.
High-content images of cells stained with orange fluorescent mitochondria dye and green fluorescent caspase dye in 2 cell types with and without treatment to induce apoptosis. Also shows quantitative dose response curves of treated cells with decreasing orange fluorescence and increasing green fluorescence at higher concentrations of drug.
Figure 5. Mitochondria membrane potential quantification using MitoTracker Orange CMTMRos. A549 or HeLa cells were cultured in Nunclon Sphera plates in GIBCO minimal essential media (MEM) to form spheroids over 2 days. Samples were treated with either DMSO vehicle or an increasing concentration of niclosamide for 24 hours before labeling with 250 nM MitoTracker Orange and 2.5 uM CellEvent Caspase 3/7 Green Reagent for 30 minutes at 37°C under normal cell culture conditions. Confocal images were acquired using the Thermo Scientific CellInsight CX7 LZR High Content Imaging System, followed by image analysis using HCS Studio Software V 2.0. Results show a niclosamide dose-dependent loss of mitochondrial membrane potential (orange) and increase in apoptotic cell death (green), qualitatively (upper panels) and quantitatively as a dose response vs mean signal intensity in both channels (lower graphs).
Kits
 Mitochondrial Membrane Potential Apoptosis KitsHCS Mitochondrial Health Kit
Apoptosis Reagent TypeMitochondrial membrane potentialMembrane Asymmetry StainMitochondrial membrane potentialCondensed chromatin stain
Reagent
MitoTracker RedAnnexin V, Alexa Fluor 488MitoHealth StainHoechst 33342
Ex/Em (nm)579/599499/521550/580350/461
Additional reagents in kitAnnexin binding buffer (5x)Image-iT DEAD Green viability stain (Ex/Em: 488/515 nm) DMSO
Size50 assays2 x 96-well plates
Cat. No.V35116H10295
High content mitochondrial health assay
The HCS Mitochondrial Health Kit was developed for simultaneous quantitative measurements of two cell health parameters by high content analysis in the same cell: mitotoxicity and cytotoxicity in a 96-well plate format. The MitoHealth stain accumulates in mitochondria in live cells proportional to the mitochondrial membrane potential (Figure 6). Cytotoxicity is measured with the Image-iT DEAD Green viability stain. The Image-iT DEAD Green viability stain has a high affinity for DNA and forms highly fluorescent and stable dye-nucleic acid complexes; it is non-fluorescent when not bound to DNA.
9 panel image showing  HeLa cells (untreated, treated with 165 nm or 120 μM valinomycin) stained with one of the following: Hoechst 33342, a cell membrane permeability stain or a mitochondrial membrane potential stain.
Figure 6. Imaging of mitotoxicity and cytotoxicity of valinomycin in HeLa cells using the HCS Mitochondrial Health Kit.
TOP
Detection of mitochondrial superoxide production
Increases in cellular superoxide production have been implicated in multiple disease states (7). This increase, which is generated as a byproduct of oxidative phosphorylation, provides another method to assess the cells’ apoptotic state.
What can be identified?
Irreversible detection of mitochondrial superoxide production in live cells.
What is the basis of the assay?
Measure the increasing fluorescence of a superoxide detection reagent that accumulates in mitochondria in a membrane potential-dependent fashion where it is selectively oxidized and subsequently binds nucleic acids.
 MitoSOX Red ReagentMitoSOX Green Reagent
Readout
  • Fluorescence increases with increased superoxide concentration
  • Highly selective detection of superoxide in the mitochondria of live cells
  • Note that two excitation peaks may be observed for MitoSOX Red dye, as it is excited at both 510 nm and 396 nm. While 510 nm will excite the superoxide oxidation product, it can also excite non-specific products, so we recommend using a 396 nm excitation for more selective detection of mitochondrial superoxide when using MitoSOX Red dye.
Ex/Em (nm)396/610488/510
Suggested filterCustom Filter, UV/Violet excitation filter and RFP Emission FilterFITC
Instrument platformFluorescence microscopy
Flow cytometry
Microplate reader
High content analysis
Fluorescence microscopy
High content analysis
Sample typeLive cellsLive cells
Fixation compatibilityNot compatible with fixation or detergentNot compatible with fixation or detergent
FormatSolid, 10 x 50 μgSolid, 10 x 50 μg
See all MitoSOX Reagents
MitoSOX Red and Green Mitochondrial Superoxide Indicators
MitoSOX Red and Green mitochondrial superoxide indicators are fluorogenic reagents designed for highly selective detection of superoxide in the mitochondria of live, healthy cells (Figure 7 and 8). The reagents are readily oxidized by superoxide but not by other reactive oxygen species (ROS)—or reactive nitrogen species (RNS)–generating systems, and oxidation of the probe is prevented by superoxide dismutase (Figures 9 and 10). Oxidation of the MitoSOX Red indicators by superoxide results in a fluorescence excitation peak at ~400 nm that is absent in the excitation spectrum generated by reactive oxygen species other than superoxide. Thus, fluorescence excitation at 400 nm with emission detection at ~590 nm provides optimum discrimination of superoxide from other reactive oxygen species. MitoSOX Green indicator can be detected with traditional FITC/GFP filter set. For selective detection of superoxide occurs with 488 nm excitation (emission at 510 nm).
Microscopic view of cells with red (mitochondria) and blue (nucleus) fluorescence in absence of treatment compared to just blue fluorescence when cells are treated with scavenger.
 Click image to enlarge
Figure 7. Imaging detection of superoxide in live cells using MitoSOX Red superoxide indicator. Live 3T3 cells were treated with FeTCPP, a superoxide scavenger, (left) or left untreated (right). Cells were then labeled with MitoSOX Red reagent, which fluoresces red when oxidized by superoxide, and nuclei were stained with blue-fluorescent Hoechst 33342. The mitochondria of untreated cells exhibited red fluorescence, indicating the presence of superoxide, whereas the mitochondria of treated cells showed minimal fluorescence.
Co-stained TMRM and MitoSOX green show co-localization when superoxide production is induced
Figure 8. Mitochondrial superoxide detection in live U2OS cells. Cells were treated overnight with 30 µM MitoPQ to induce superoxide production (A) or with ethanol as vehicle control (B). Cells were then stained with 1 µM MitoSOX Green and 100 nM TMRM for 30 minutes to label mitochondria. Nuclei were labeled with Hoechst 33342 and then imaged in HBSS on Zeiss confocal.
Graphs showing very low reactivity of both MitoSOX Green and Red with other ROS or NOS species as indicated by relative integrated fluorescence.
Graphs showing very low reactivity of both MitoSOX Green and Red with other ROS or NOS species as indicated by relative integrated fluorescence.
Figure 9. Selectivity of MitoSOX Green (MSG) and MitoSOX Red (MSR) mitochondrial superoxide indicators in a cell free environment. Cell free systems were used to generate reactive oxygen species (ROS). Each ROS was added to separate solutions of 10 µM MSG and MSR and incubated for 30 min at ambient temperature. Excess DNA was added to the solutions with MSR. Superoxide dismutase was used as a negative control for superoxide. Superoxide was generated using potassium superoxide for MSG, and the xanthine/xanthine oxidase system was used to generate superoxide for MSR. The results show that both MSG and MSR are selective to superoxide and not other reactive oxygen species.
Multi-panel figure with microscopic views of cells untreated with high glucose medium that are stained with red and blue fluorescence compared to low glucose with untreated cells and cells treated with the nitric oxide generator showing blue fluorescence only, and cells treated with superoxide generator stained with both red and blue fluorescence.
Figure 10. Visualizing glucose-mediated oxidative stress. Live human osteosarcoma (U2OS) cells were plated in Minimum Essential Medium (MEM) and incubated overnight at 37°C with CO2. To mitigate the effect of high glucose–mediated oxidative stress, samples B–D were washed in PBS and immersed in low-glucose Dulbecco’s Modified Eagle Medium (DMEM) and incubated overnight at 37°C with CO2. Samples were then washed in Hanks' Balanced Salt Solution (HBSS) and then treated as follows for the next 30 minutes: (A, B) cells were left in HBSS; (C) cells were incubated in HBSS + 100 μM antimycin A; (D) cells were incubated in HBSS + 100 μM DEANO. MitoSOX Red reagent at 5 µM was added to each sample, and cells were incubated for 30 minutes and imaged by confocal microscopy.
Detection of mitochondrial calcium
What can be identified?
Irreversible detection of mitochondrial calcium in live cells.
What is the basis of the assay?
Measure the increasing fluorescence of a calcium indicator with increasing calcium concentration in the mitochondria.
 Rhod-2, AM reagent
Readout
  • Calcium indicator targeted to mitochondria
  • Increases fluorescence with Ca 2+ concentration
Ex/Em (nm)552/577
Suggested filterTRITC
Instrument platformFluorescence microscopy
Flow cytometry
Sample typeLive cells
Fixation compatibilityNot compatible with fixation or detergent
FormatSolid, 20 x 50 mg
Cat. No.R1245MP
Rhod-2, AM Reagent
Elevated mitochondrial Ca2+ plays an important role in initiation of programmed cell death (apoptosis) as well as in other cellular processes (8). Fluorescent probes that show a spectral response upon binding Ca2+ have enabled researchers to investigate changes in intracellular free Ca2+ concentrations using fluorescence microscopy, flow cytometry and fluorescence spectroscopy. As shown in Figure 11, Rhod-2 is a fluorescent calcium indicator that can also be used to detect mitochondrial Ca2+ (8). The AM ester form of Rhod-2 (Rhod-2, AM) is used to easily load the dye into live cells. Upon entering the cell, intracellular esterases cleave the AM group, freeing the Rhod-2 salt form to bind and fluoresce upon binding to mitochondrial Ca2+.
4 panels showing microscopic view of cells stained with green fluorescent mitochondria-targeting GFP and orange fluorescence of rhod-2 calcium indicator, also localized in mitochondria.
Figure 11. Multiplex imaging of mitochondrial calcium levels and dynamics.(A) HeLa cells were labeled with CellLight Mitochondria-GFP and 5 μM Rhod-2, AM for 15 min at 37°C before imaging live over 100 sec. (B–D) The region outlined in (A) is enlarged to show individual mitochondria within a single cell over time. (C, D) Calcium is released from internal stores following application of 10 μM histamine. Mitochondria in close proximity to the calcium release are revealed by the increase in the orange-red fluorescence of Rhod-2. The arrow in (C) denotes a mitochondrion that may have impaired calcium uptake, a detail that would have been missed using Rhod-2, AM alone. The asterisk marks a mitochondrion that shows a transient elevation in calcium levels.
Detection of changes in mitochondrial permeability transition pore
What can be identified?
Irreversible detection of the mitochondrial permeability transition (MPT) subsequent to dysregulation of the MPT pore.
What is the basis of the assay?
Loss of mitochondrial calcein fluorescence due to mitochondrial permeability transition pore opening.
 Image-IT LIVE Mitochondrial Transition Pore AssayMitoProbes Transition Pore Assay Kit
ApplicationMethod of measuring mitochondrial permeability transition pore opening
ReadoutIn healthy cells, the mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.
Ex/Em (nm)494/517 (calcein)494/517 (calcein)
579/599 (MitoTracker Red stain)
361/497 (Hoechst 33342)
Suggested filterTRITCFITC
Instrument platformFluorescence microscopyFlow cytometry
Sample typeLive cellsLive cells
Compatibility with fixationNot compatible with fixation and detergentNot compatible with fixation and detergent
Kit componentsKit contents:
Calcein AM (calcium indicator)
Ionomycin (ionophore)
CoCl2 (calcein quencher)
MitoTracker Red CMXRos stain
Hoechst 33342 (nuclear stain)
Kit contents:
Calcein AM (calcium indicator)
Ionomycin (ionophore)
CoCl2 (calcein quencher)
DMSO
Cat. No.I35103M34153
Mitochondrial permeability transition pore assays
The mitochondrial permeability transition pore (MPTP) is a nonspecific channel formed by components from the inner and outer mitochondrial membranes, and appears to be involved in the release of mitochondrial components during cell death. This opening of the pore dramatically alters the permeability of mitochondria, as well as the mitochondria membrane potential. This continuous pore activation results from mitochondrial Ca2+ overload, oxidation of mitochondrial glutathione, increased levels of reactive oxygen species in mitochondria, and other pro-apoptotic conditions.
We have developed two kits for detecting mitochondrial transition pore opening, one for imaging microscopy (Image-iT LIVE Mitochondrial Transition Pore Assay Kit, Figure 12) and the other for flow cytometry (MitoProbe Transition Pore Assay Kit, Figure 13). Both kits provide a more direct method of measuring mitochondrial permeability transition pore opening than assays relying on mitochondrial membrane potential alone. This assay employs the acetoxymethyl (AM) ester of calcein, a colorless and nonfluorescent esterase substrate, and CoCl2 (cobalt), a quencher of calcein fluorescence, to selectively label mitochondria. Cells are loaded with calcein AM, which passively diffuses into the cells and accumulates in cytosolic compartments, including the mitochondria. Once inside cells, calcein AM is cleaved by intracellular esterases to liberate the very polar fluorescent dye calcein, which does not cross the mitochondrial or plasma membranes in appreciable amounts over relatively short periods of time. The fluorescence from cytosolic calcein is quenched by the addition of CoCl2, while the fluorescence from the mitochondrial calcein is maintained. As a control, cells that have been loaded with calcein AM and CoCl2 can also be treated with a Ca2+ ionophore such as ionomycin to allow entry of excess Ca2+ into the cells, which triggers mitochondrial pore activation and subsequent loss of mitochondrial calcein fluorescence. This ionomycin response can be blocked with cyclosporin A, a compound reported to prevent mitochondrial transition pore formation by binding cyclophilin D.
Figure 12. BPAE cells stained with Image-iT LIVE Mitochondrial Transition Pore Assay Kit. BPAE cells are stained using components from the Image-iT LIVE Mitochondrial Transition Pore Assay Kit. Cells are counterstained with MitoTracker Red CMXRos to show mitochondria and with Hoechst 33342 to show nuclei. Panel A shows the uniform cellular fluorescence from unquenched calcein. Panel B shows the mitochondrial pattern after adding cobalt which quenches the cytoplasmic calcein fluorescence but not mitochondrial calcein. Panel C shows the loss of calcein fluorescence with the addition of ionomycin which opens the pore to allow cobalt in and calcein out (mitochondria still visible from MitoTracker Red). Cyclophilin D activity is necessary for MPTP formation, and is inhibited by cyclosporin A. Consequently, inhibition of pore formation by cyclosporin A (CspA) has been used as an argument that a function is MPTP-specific. Panel D shows that the calcein pattern is retained when cyclosporin A is added before ionomycin, indicating that the ionomycin-triggered change observed here is an MPTP-mediated event.
3 flow cytometry histograms showing shift in number of cells with green fluorescence, from high levels in untreated cells, mid levels when mitochondrial calcein is quenched, down to very low levels after fluorescence signal is quenched in both cytosol and mitochondria.
Figure 13. MitoProbe Transition Pore Assay Kit for flow cytometry. The flow cytometry histograms show the actions of the various kit components. Jurkat cells were incubated with the reagents in the MitoProbe Transition Pore Assay Kit and analyzed by flow cytometry. (A) In the absence of CoCl2 and ionomycin, fluorescent calcein is present in the cytosol as well as the mitochondria, resulting in a bright signal. (B) In the presence of CoCl2, calcein in the mitochondria emits a signal, but the cytosolic calcein fluorescence is quenched the overall fluorescence is reduced compared to calcein alone. (C) When ionomycin, a calcium ionophore, and CoCl2 are added to the cells at the same time as calcein AM, the fluorescence signals from both the cytosol and mitochondria are largely abolished.
TOP
Mitochondrial function changes in apoptosis
A feature of the early stages of programmed cell death is the disruption of mitochondrial function. This mitochondrial disruption includes changes in the membrane potential - a central feature of mitochondrial health, and alterations to the oxidation–reduction potential of the mitochondria. The inner mitochondrial membrane potential is essential in Ca2+ uptake and storage, reactive oxygen species generation, detoxification, and most importantly, the synthesis of ATP by oxidative phosphorylation (1). Therefore, the membrane’s depolarization is a good indicator of mitochondrial dysfunction, which is increasingly implicated in drug toxicity (2-6). Changes in the membrane potential, along with decreases ATP to ADP ratios, increased mitochondrial matrix calcium levels, oxidative stress, and release of cytochrome c into the cytosol, are all presumed to be associated with the mitochondrial permeability transition, resulting in disruption of ions and small molecule homeostasis via the mitochondrial permeability transition pore (MPTP).
Mitochondria function assays
The disruption of mitochondria function can be detected using a variety of fluorescence-based assays including measurements of mitochondrial calcium, superoxide, mitochondrial permeability transition, and membrane potential. A summary of these assays are listed in Table 1 below.
Although disruption of mitochondrial function is associated with apoptosis, the various aspects listed above are not specific to apoptotic cell death. Because the aforementioned apoptosis-related mitochondria function parameters are not specific for apoptosis and may vary across cell types, it is often advantageous to consider multi-parametric assays, including the use of specific markers for the activation of particular caspase proteases.
Summary of mitochondria function assays
 Mitochondria function assays
 Membrane potential and MetabolismSuperoxide productionCalciumMitochondrial permeability transition
What can be identified?
Cells with healthy, metabolically active mitochondria.
2 options available:
Detection of dynamic changes in relative mitochondria membrane potential (reversible detection).
End point assays for detecting relative mitochondria membrane potential at a specific timepoint (irreversible detection).
Irreversible detection of mitochondrial superoxide production in live cells.
Irreversible detection of mitochondrial calcium in live cells.Irreversible detection of the mitochondrial permeability transition (MPT) subsequent to dysregulation of the MPT pore.
What is the basis of assay?Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.Measure the increasing fluorescence of a superoxide detection reagent that accumulates in mitochondria in a membrane potential-dependent fashion where it is selectively oxidized and subsequently binds nucleic acids.Measure the increasing fluorescence of a calcium indicator with increasing calcium concentration in the mitochondria.Loss of mitochondrial calcein fluorescence due to mitochondrial permeability transition.
Is the assay fixable?
Only a small selection of the end point assays for detecting mitochondria membrane potential are fixable. Please see the table of MitoTracker probes for Imaging and Flow Cytometry below.
None of the dynamic assays are fixable.
Not compatible with fixation.
Not compatible with fixation.
Not compatible with fixation.
Detection of dynamic changes in mitochondrial membrane potential
What can be identified?
Cells with healthy, metabolically active mitochondria. Dynamic changes detect relative mitochondria membrane potential (reversible detection). None of the dynamic assays are fixable.
What is the basis of the assay?
Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.
Dual-emission, ratiometric reagents
 JC-1MitoProbe JC-1 Assay KitMitoProbe DiOC2(3) Assay Kit
ReadoutActive mitochondria exhibit brighter red fluorescence signal compared to mitochondria with lower membrane potential which fluoresce green. 
Ex/Em (nm)514/529 (monomer, green)485/497 (monomer, green)
514/590 (aggregate, red)485/650 (aggregate, far–red)
Suggested filtersTRITCFITC and PEFITC and APC
Instrument platformImaging microscopyFlow cytometryFlow cytometry
Sample typeLive cellsLive cellsLive cells
Compatibility with fixationNoNoNo
Format5 mgKit contents:
JC-1, DMSO
CCCP (a mitochondria membrane potential disrupter in DMSO)
10x PBS
Kit contents:
DiOC2(3) in DMSO
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Cat. No.T3168M34152M34150
Single-emission reagents
 TMRMImage-iT TMRMMitoProbe TMRM Assay KitMitoProbe DiIC1(5) Assay Kit
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)548/574638/658
Suggested filterTRITC~585/16 nmAlexa Fluor 647/APC
Instrument platformFluorescence microscopyFlow cytometry
Sample typeLive cellsLive cellsLive cellsLive cells
Compatibility with fixationNoNoNoNo
Format25 mg5 x 100 μLKit contents:
TMRM
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Kit contents:
DiIC1(5) in DMSO
CCCP (a mitochondrial membrane potential disrupter in DMSO)
Cat. No.T668I34361M20036M34151
JC-1 Dye and the MitoProbe JC-1 Assay Kit
JC-1 is a cell-permeant dye widely used for detecting mitochondrial membrane potential in flow cytometry and microscopy platforms (Figure 1). JC-1 exhibits potential-dependent accumulate within the mitochondria, forming J-aggregates as the dye increases. This increase in potential will result in a shift from the green fluorescence (~529 nm) monomer to red fluorescence (~590 nm) J-aggregates. Alternatively, depolarization of the mitochondria is denoted by a decrease in the red/green fluorescence intensity ratio. 
Learn more about the JC-1 Dye for mitochondria membrane potential
A
Microscopic view of mitochondria stained with green-red fluorescence at 4 different time course points.
B
Dot plot of JC-1 stained cells showing 2 cell populations-healthy cells colored red and apoptotic cells colored green.
Figure 1. Detection of mitochondria membrane potential using JC-1 Dye.(A) NIH 3T3 fibroblasts stained with JC-1 Dye show the progressive loss of red J-aggregate fluorescence and cytoplasmic diffusion of green monomer fluorescence following exposure to hydrogen peroxide. Images show the same field of cells viewed before H2O2 treatment and 5, 10 and 20 min after treatment. The images were contributed by Ildo Nicoletti, Perugia University Medical School. (B)MitoProbe JC-1 Assay Kit was used to stain Jurkat cells (T-cell leukemia, human) which were then analyzed on a flow cytometer using 488 nm excitation with 530 nm and 585 nm bandpass emission filters. Green = apoptotic cells (reduced mitochondria membrane potential), red = normal cells.
MitoProbe DiOC2(3) Assay Kit
Another dye that can be used as a ratiometric probe for mitochondria membrane potential is DiOC2(3). The mechanism is similar to that of JC-1 Dye in that the monomer exhibits green fluorescence at lower concentrations where mitochondria membrane potential is low and the aggregate of the dye in more active mitochondria causes the emission to shift toward the red. In the case of DiOC2(3), the shift is farther in the red than JC-1, above 650 nm. The MitoProbe DiOC2(3) Assay Kit has been specifically developed to work in flow cytometry applications and includes a mitochondrial membrane-potential disrupter, CCCP.
TOP
TMRM and MitoProbe TMRM Assay Kit
Tetramethylrhodamine, methyl ester (TMRM) or the related TMRE (tetramethylrhodamine, ethyl ester) are small, cell-permeant dyes that accumulate in active mitochondria. If the cells are healthy and have functioning mitochondria, the signal is bright. Upon loss of the mitochondrial membrane potential, TMRM and TMRE accumulation ceases and the signal dims or is lost from the mitochondria. This ability to dynamically monitor changes in mitochondria membrane potential is a distinct advantage of this probe. TMRM and TMRE signals can be detected in live cells or isolate mitochondria with traditional and high content fluorescence microscopy, microplates, or by flow cytometry (Figure 2).
TMRM is available in different formats to fit your apoptosis assay needs. See the Selection Guide (Tab 2. Single-emission reagents) for detection of dynamic changes in mitochondria membrane potential using single-emission reagents. MitoProbe TMRM Assay Kit for Flow Cytometry includes TMRM along with CCCP, a mitochondrial membrane potential disrupter and detailed protocol for use in flow cytometry.
A
Microscopic view of cells stained with orange fluorescence (mitochondria) and green fluorescence (tubulin)
B
Dot plot showing healthy, apoptotic and dead cells with and without treatment.
Figure 2. TMRM as a mitochondrial membrane potential assay for apoptosis.(A) HeLa cells labeled using Tubulin Tracker Green and Image-iT TMRM Reagent show superb multiplexing capability and staining specificity. Image shows multiple live mitotic cells with microtubules assembled into a mitotic spindle with visible microtubule filaments, as well as cleavage furrow indicating completion of cytokinesis. (B) Jurkat cells, a human T-lymphocyte cell line, were treated with DMSO (control) or 500 nM staurosporine for 2 hours. Cells were subsequently stained with MitoProbe TMRM for 30 min at 37˚C, followed by a wash and additional stain with Annexin V Pacific Blue conjugate. Staurosporine induced apoptosis, resulting in a mixed population of cells containing a population of healthy MitoProbe TMRM-positive cells as well as a population of apoptotic, Annexin V Pacific Blue-positive, MitoProbe TMRM-low cells.
TOP
MitoProbe DiIC1(5) dye
Cationic carbocyanine dyes have been shown to accumulate in cells in response to membrane potential. The MitoProbe DiIC1(5) Kit is designed for flow cytometry applications and provides the far-red–fluorescent DiIC1(5) carbocyanine dye, along with a mitochondrial membrane potential disrupter, CCCP. At concentrations below 100 nm, the cationic DiIC1(5) dye passively accumulates in the mitochondria with active membrane potentials in living cells (Figure 3). Unlike DiOC2(3), however, DiIC1(5) dye exhibits a single wavelength emission whose fluorescence signal increases with more active mitochondria.
Flow cytometry histogram showing cells with active mitochondria in control compared to cells with depolarized mitochondria with CCCP treatment
Figure 3. Detection of changes in mitochondrial membrane potential using the MitoProbe DiIC1(5) Assay Kit for Flow Cytometry. Decrease in mitochondrial membrane potential as demonstrated with DiIC1(5) fluorescence due to the addition of carbonyl cyanide 3-chlorophenylhydrazone (CCCP). Jurkat cells (T-cell leukemia, human) were stained with 50 nM DiIC1(5) alone (blue) or in the presence of 50 μM CCCP (red).
TOP
End point assays in mitochondrial membrane potential
What can be identified?
Cells with healthy, metabolically active mitochondria. End point assays detect relative mitochondria membrane potential at a specific timepoint (irreversible detection).
Only a small selection of the end point assays for detecting mitochondria membrane potential are fixable.
What is the basis of the assay?
Measure fluorescence of reagents that accumulate in active (healthy) mitochondria with intact membrane potentials.
 MitoTracker Probes—Imaging
MitoTracker Green FMMitoTracker Orange CMTMRosMitoTracker Red CMXRosMitoTracker Red FMMitoTracker Deep Red FM
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)490/516550/580579/599581/644644/665
Suggested filterFITCTRITCTexas RedCy5Cy5
Instrument platformFluorescence microscopy
Microplate reader
Sample typeLive cellsLive cellsLive cellsLive cellsLive cells
Compatibility with fixation NoYesYesNoNo
Format20 x 50 μg20 x 50 μg20 x 50 μg20 x 50 μg20 x 50 μg
See all MitoTracker dyes
 MitoTracker Probes—Flow Cytometry
MitoTracker Green FMMitoTracker Red CMXRosMitoTracker Red FMMitoTracker Deep Red FM
ReadoutActive mitochondria exhibit brighter fluorescence compared to apoptotic mitochondria.
Ex/Em (nm)490/516579/599581/644644/665
Suggested filterFITCTexas RedCy5Cy5
Instrument platformFlow cytometry
Sample typeLive cellsLive cellsLive cellsLive cells
Compatibility with fixation*NoYesNoNo
Format5 x 20 μg5 x 20 μg5 x 20 μg5 x 20 μg
Cat. No.M7514M46752M46751M46753
MitoTracker probes
MitoTracker probes are small (<1 kDa), cell-permeant mitochondrion-selective dyes that contain a thiol-reactive chloromethyl moiety that keeps the dye associated with the mitochondria after fixation. Because the probes form covalent bonds with the mitochondrial thiols, they can only be used as an end point assays to detect mitochondrial membrane potential at the time of loading in live cells, and should not be used as sensors of dynamic mitochondrial membrane changes over time.
Impact of fixation on MitoTracker probes
While the MitoTracker signal can be fixed after labeling, in assays to study mitochondrial structure, if studying the mitochondrial membrane function, it is strongly suggested to use a few, select MitoProbe reagents. Only MitoTracker Orange CMTMRos and MitoTracker Red CMXRos have been found to maintain a consistent difference in signal loss when comparing mitochondrial membrane potential in control and treated samples after fixation. These single wavelength emission reagents can be used for flow cytometry, imaging microscopy, and high content analysis applications (Figures 4 and 5).
Learn more about MitoTracker probes for mitochondria structure
Flow cytometry dot plot showing separation of apoptotic and healthy cells in treated versus untreated cells.
 Click image to enlarge
Figure 4. MitoTracker Red CMXRos dye mitochondria membrane potential detection. Jurkat human T-cell leukemia cells in complete medium were (A) first exposed to 10 µM camptothecin for 4 hours or (B) left untreated. Both cell populations were then treated with the reagents in the Mitochondrial Membrane Potential/Annexin V Apoptosis Kit and analyzed by flow cytometry. Note that the apoptotic cells show higher reactivity for annexin V and lower MitoTracker Red dye fluorescence than do live cells.
High-content images of cells stained with orange fluorescent mitochondria dye and green fluorescent caspase dye in 2 cell types with and without treatment to induce apoptosis. Also shows quantitative dose response curves of treated cells with decreasing orange fluorescence and increasing green fluorescence at higher concentrations of drug.
Figure 5. Mitochondria membrane potential quantification using MitoTracker Orange CMTMRos. A549 or HeLa cells were cultured in Nunclon Sphera plates in GIBCO minimal essential media (MEM) to form spheroids over 2 days. Samples were treated with either DMSO vehicle or an increasing concentration of niclosamide for 24 hours before labeling with 250 nM MitoTracker Orange and 2.5 uM CellEvent Caspase 3/7 Green Reagent for 30 minutes at 37°C under normal cell culture conditions. Confocal images were acquired using the Thermo Scientific CellInsight CX7 LZR High Content Imaging System, followed by image analysis using HCS Studio Software V 2.0. Results show a niclosamide dose-dependent loss of mitochondrial membrane potential (orange) and increase in apoptotic cell death (green), qualitatively (upper panels) and quantitatively as a dose response vs mean signal intensity in both channels (lower graphs).
Kits
 Mitochondrial Membrane Potential Apoptosis KitsHCS Mitochondrial Health Kit
Apoptosis Reagent TypeMitochondrial membrane potentialMembrane Asymmetry StainMitochondrial membrane potentialCondensed chromatin stain
Reagent
MitoTracker RedAnnexin V, Alexa Fluor 488MitoHealth StainHoechst 33342
Ex/Em (nm)579/599499/521550/580350/461
Additional reagents in kitAnnexin binding buffer (5x)Image-iT DEAD Green viability stain (Ex/Em: 488/515 nm) DMSO
Size50 assays2 x 96-well plates
Cat. No.V35116H10295
High content mitochondrial health assay
The HCS Mitochondrial Health Kit was developed for simultaneous quantitative measurements of two cell health parameters by high content analysis in the same cell: mitotoxicity and cytotoxicity in a 96-well plate format. The MitoHealth stain accumulates in mitochondria in live cells proportional to the mitochondrial membrane potential (Figure 6). Cytotoxicity is measured with the Image-iT DEAD Green viability stain. The Image-iT DEAD Green viability stain has a high affinity for DNA and forms highly fluorescent and stable dye-nucleic acid complexes; it is non-fluorescent when not bound to DNA.
9 panel image showing  HeLa cells (untreated, treated with 165 nm or 120 μM valinomycin) stained with one of the following: Hoechst 33342, a cell membrane permeability stain or a mitochondrial membrane potential stain.
Figure 6. Imaging of mitotoxicity and cytotoxicity of valinomycin in HeLa cells using the HCS Mitochondrial Health Kit.
TOP
Detection of mitochondrial superoxide production
Increases in cellular superoxide production have been implicated in multiple disease states (7). This increase, which is generated as a byproduct of oxidative phosphorylation, provides another method to assess the cells’ apoptotic state.
What can be identified?
Irreversible detection of mitochondrial superoxide production in live cells.
What is the basis of the assay?
Measure the increasing fluorescence of a superoxide detection reagent that accumulates in mitochondria in a membrane potential-dependent fashion where it is selectively oxidized and subsequently binds nucleic acids.
 MitoSOX Red ReagentMitoSOX Green Reagent
Readout
  • Fluorescence increases with increased superoxide concentration
  • Highly selective detection of superoxide in the mitochondria of live cells
  • Note that two excitation peaks may be observed for MitoSOX Red dye, as it is excited at both 510 nm and 396 nm. While 510 nm will excite the superoxide oxidation product, it can also excite non-specific products, so we recommend using a 396 nm excitation for more selective detection of mitochondrial superoxide when using MitoSOX Red dye.
Ex/Em (nm)396/610488/510
Suggested filterCustom Filter, UV/Violet excitation filter and RFP Emission FilterFITC
Instrument platformFluorescence microscopy
Flow cytometry
Microplate reader
High content analysis
Fluorescence microscopy
High content analysis
Sample typeLive cellsLive cells
Fixation compatibilityNot compatible with fixation or detergentNot compatible with fixation or detergent
FormatSolid, 10 x 50 μgSolid, 10 x 50 μg
See all MitoSOX Reagents
MitoSOX Red and Green Mitochondrial Superoxide Indicators
MitoSOX Red and Green mitochondrial superoxide indicators are fluorogenic reagents designed for highly selective detection of superoxide in the mitochondria of live, healthy cells (Figure 7 and 8). The reagents are readily oxidized by superoxide but not by other reactive oxygen species (ROS)—or reactive nitrogen species (RNS)–generating systems, and oxidation of the probe is prevented by superoxide dismutase (Figures 9 and 10). Oxidation of the MitoSOX Red indicators by superoxide results in a fluorescence excitation peak at ~400 nm that is absent in the excitation spectrum generated by reactive oxygen species other than superoxide. Thus, fluorescence excitation at 400 nm with emission detection at ~590 nm provides optimum discrimination of superoxide from other reactive oxygen species. MitoSOX Green indicator can be detected with traditional FITC/GFP filter set. For selective detection of superoxide occurs with 488 nm excitation (emission at 510 nm).
Microscopic view of cells with red (mitochondria) and blue (nucleus) fluorescence in absence of treatment compared to just blue fluorescence when cells are treated with scavenger.
 Click image to enlarge
Figure 7. Imaging detection of superoxide in live cells using MitoSOX Red superoxide indicator. Live 3T3 cells were treated with FeTCPP, a superoxide scavenger, (left) or left untreated (right). Cells were then labeled with MitoSOX Red reagent, which fluoresces red when oxidized by superoxide, and nuclei were stained with blue-fluorescent Hoechst 33342. The mitochondria of untreated cells exhibited red fluorescence, indicating the presence of superoxide, whereas the mitochondria of treated cells showed minimal fluorescence.
Co-stained TMRM and MitoSOX green show co-localization when superoxide production is induced
Figure 8. Mitochondrial superoxide detection in live U2OS cells. Cells were treated overnight with 30 µM MitoPQ to induce superoxide production (A) or with ethanol as vehicle control (B). Cells were then stained with 1 µM MitoSOX Green and 100 nM TMRM for 30 minutes to label mitochondria. Nuclei were labeled with Hoechst 33342 and then imaged in HBSS on Zeiss confocal.
Graphs showing very low reactivity of both MitoSOX Green and Red with other ROS or NOS species as indicated by relative integrated fluorescence.
Graphs showing very low reactivity of both MitoSOX Green and Red with other ROS or NOS species as indicated by relative integrated fluorescence.
Figure 9. Selectivity of MitoSOX Green (MSG) and MitoSOX Red (MSR) mitochondrial superoxide indicators in a cell free environment. Cell free systems were used to generate reactive oxygen species (ROS). Each ROS was added to separate solutions of 10 µM MSG and MSR and incubated for 30 min at ambient temperature. Excess DNA was added to the solutions with MSR. Superoxide dismutase was used as a negative control for superoxide. Superoxide was generated using potassium superoxide for MSG, and the xanthine/xanthine oxidase system was used to generate superoxide for MSR. The results show that both MSG and MSR are selective to superoxide and not other reactive oxygen species.
Multi-panel figure with microscopic views of cells untreated with high glucose medium that are stained with red and blue fluorescence compared to low glucose with untreated cells and cells treated with the nitric oxide generator showing blue fluorescence only, and cells treated with superoxide generator stained with both red and blue fluorescence.
Figure 10. Visualizing glucose-mediated oxidative stress. Live human osteosarcoma (U2OS) cells were plated in Minimum Essential Medium (MEM) and incubated overnight at 37°C with CO2. To mitigate the effect of high glucose–mediated oxidative stress, samples B–D were washed in PBS and immersed in low-glucose Dulbecco’s Modified Eagle Medium (DMEM) and incubated overnight at 37°C with CO2. Samples were then washed in Hanks' Balanced Salt Solution (HBSS) and then treated as follows for the next 30 minutes: (A, B) cells were left in HBSS; (C) cells were incubated in HBSS + 100 μM antimycin A; (D) cells were incubated in HBSS + 100 μM DEANO. MitoSOX Red reagent at 5 µM was added to each sample, and cells were incubated for 30 minutes and imaged by confocal microscopy.
Detection of mitochondrial calcium
What can be identified?
Irreversible detection of mitochondrial calcium in live cells.
What is the basis of the assay?
Measure the increasing fluorescence of a calcium indicator with increasing calcium concentration in the mitochondria.
 Rhod-2, AM reagent
Readout
  • Calcium indicator targeted to mitochondria
  • Increases fluorescence with Ca 2+ concentration
Ex/Em (nm)552/577
Suggested filterTRITC
Instrument platformFluorescence microscopy
Flow cytometry
Sample typeLive cells
Fixation compatibilityNot compatible with fixation or detergent
FormatSolid, 20 x 50 mg
Cat. No.R1245MP
Rhod-2, AM Reagent
Elevated mitochondrial Ca2+ plays an important role in initiation of programmed cell death (apoptosis) as well as in other cellular processes (8). Fluorescent probes that show a spectral response upon binding Ca2+ have enabled researchers to investigate changes in intracellular free Ca2+ concentrations using fluorescence microscopy, flow cytometry and fluorescence spectroscopy. As shown in Figure 11, Rhod-2 is a fluorescent calcium indicator that can also be used to detect mitochondrial Ca2+ (8). The AM ester form of Rhod-2 (Rhod-2, AM) is used to easily load the dye into live cells. Upon entering the cell, intracellular esterases cleave the AM group, freeing the Rhod-2 salt form to bind and fluoresce upon binding to mitochondrial Ca2+.
4 panels showing microscopic view of cells stained with green fluorescent mitochondria-targeting GFP and orange fluorescence of rhod-2 calcium indicator, also localized in mitochondria.
Figure 11. Multiplex imaging of mitochondrial calcium levels and dynamics.(A) HeLa cells were labeled with CellLight Mitochondria-GFP and 5 μM Rhod-2, AM for 15 min at 37°C before imaging live over 100 sec. (B–D) The region outlined in (A) is enlarged to show individual mitochondria within a single cell over time. (C, D) Calcium is released from internal stores following application of 10 μM histamine. Mitochondria in close proximity to the calcium release are revealed by the increase in the orange-red fluorescence of Rhod-2. The arrow in (C) denotes a mitochondrion that may have impaired calcium uptake, a detail that would have been missed using Rhod-2, AM alone. The asterisk marks a mitochondrion that shows a transient elevation in calcium levels.
Detection of changes in mitochondrial permeability transition pore
What can be identified?
Irreversible detection of the mitochondrial permeability transition (MPT) subsequent to dysregulation of the MPT pore.
What is the basis of the assay?
Loss of mitochondrial calcein fluorescence due to mitochondrial permeability transition pore opening.
 Image-IT LIVE Mitochondrial Transition Pore AssayMitoProbes Transition Pore Assay Kit
ApplicationMethod of measuring mitochondrial permeability transition pore opening
ReadoutIn healthy cells, the mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.
Ex/Em (nm)494/517 (calcein)494/517 (calcein)
579/599 (MitoTracker Red stain)
361/497 (Hoechst 33342)
Suggested filterTRITCFITC
Instrument platformFluorescence microscopyFlow cytometry
Sample typeLive cellsLive cells
Compatibility with fixationNot compatible with fixation and detergentNot compatible with fixation and detergent
Kit componentsKit contents:
Calcein AM (calcium indicator)
Ionomycin (ionophore)
CoCl2 (calcein quencher)
MitoTracker Red CMXRos stain
Hoechst 33342 (nuclear stain)
Kit contents:
Calcein AM (calcium indicator)
Ionomycin (ionophore)
CoCl2 (calcein quencher)
DMSO
Cat. No.I35103M34153
Mitochondrial permeability transition pore assays
The mitochondrial permeability transition pore (MPTP) is a nonspecific channel formed by components from the inner and outer mitochondrial membranes, and appears to be involved in the release of mitochondrial components during cell death. This opening of the pore dramatically alters the permeability of mitochondria, as well as the mitochondria membrane potential. This continuous pore activation results from mitochondrial Ca2+ overload, oxidation of mitochondrial glutathione, increased levels of reactive oxygen species in mitochondria, and other pro-apoptotic conditions.
We have developed two kits for detecting mitochondrial transition pore opening, one for imaging microscopy (Image-iT LIVE Mitochondrial Transition Pore Assay Kit, Figure 12) and the other for flow cytometry (MitoProbe Transition Pore Assay Kit, Figure 13). Both kits provide a more direct method of measuring mitochondrial permeability transition pore opening than assays relying on mitochondrial membrane potential alone. This assay employs the acetoxymethyl (AM) ester of calcein, a colorless and nonfluorescent esterase substrate, and CoCl2 (cobalt), a quencher of calcein fluorescence, to selectively label mitochondria. Cells are loaded with calcein AM, which passively diffuses into the cells and accumulates in cytosolic compartments, including the mitochondria. Once inside cells, calcein AM is cleaved by intracellular esterases to liberate the very polar fluorescent dye calcein, which does not cross the mitochondrial or plasma membranes in appreciable amounts over relatively short periods of time. The fluorescence from cytosolic calcein is quenched by the addition of CoCl2, while the fluorescence from the mitochondrial calcein is maintained. As a control, cells that have been loaded with calcein AM and CoCl2 can also be treated with a Ca2+ ionophore such as ionomycin to allow entry of excess Ca2+ into the cells, which triggers mitochondrial pore activation and subsequent loss of mitochondrial calcein fluorescence. This ionomycin response can be blocked with cyclosporin A, a compound reported to prevent mitochondrial transition pore formation by binding cyclophilin D.
Figure 12. BPAE cells stained with Image-iT LIVE Mitochondrial Transition Pore Assay Kit. BPAE cells are stained using components from the Image-iT LIVE Mitochondrial Transition Pore Assay Kit. Cells are counterstained with MitoTracker Red CMXRos to show mitochondria and with Hoechst 33342 to show nuclei. Panel A shows the uniform cellular fluorescence from unquenched calcein. Panel B shows the mitochondrial pattern after adding cobalt which quenches the cytoplasmic calcein fluorescence but not mitochondrial calcein. Panel C shows the loss of calcein fluorescence with the addition of ionomycin which opens the pore to allow cobalt in and calcein out (mitochondria still visible from MitoTracker Red). Cyclophilin D activity is necessary for MPTP formation, and is inhibited by cyclosporin A. Consequently, inhibition of pore formation by cyclosporin A (CspA) has been used as an argument that a function is MPTP-specific. Panel D shows that the calcein pattern is retained when cyclosporin A is added before ionomycin, indicating that the ionomycin-triggered change observed here is an MPTP-mediated event.
3 flow cytometry histograms showing shift in number of cells with green fluorescence, from high levels in untreated cells, mid levels when mitochondrial calcein is quenched, down to very low levels after fluorescence signal is quenched in both cytosol and mitochondria.
Figure 13. MitoProbe Transition Pore Assay Kit for flow cytometry. The flow cytometry histograms show the actions of the various kit components. Jurkat cells were incubated with the reagents in the MitoProbe Transition Pore Assay Kit and analyzed by flow cytometry. (A) In the absence of CoCl2 and ionomycin, fluorescent calcein is present in the cytosol as well as the mitochondria, resulting in a bright signal. (B) In the presence of CoCl2, calcein in the mitochondria emits a signal, but the cytosolic calcein fluorescence is quenched the overall fluorescence is reduced compared to calcein alone. (C) When ionomycin, a calcium ionophore, and CoCl2 are added to the cells at the same time as calcein AM, the fluorescence signals from both the cytosol and mitochondria are largely abolished.
TOP
Ordering information
Catalog # Name Size Price (USD) Qty
R1245MP Rhod-2, AM, cell permeant, 20 x 50μg Each
669.65
Online Exclusive
736.00
Save 66.35 (9%)
M36008 MitoSOX™ Red Mitochondrial Superoxide Indicator, for live-cell imaging, 10 vials x 50 μg Each
435.65
Online Exclusive
464.00
Save 28.35 (6%)
M36006 MitoSOX™ Green Mitochondrial Superoxide Indicator, for live-cell imaging, 5 vials x 9 μg Each
442.65
Online Exclusive
471.00
Save 28.35 (6%)
Catalog # Name Size Price (USD) Qty
I35103 Image-IT™ LIVE Mitochondrial Transition Pore Assay Kit, for microscopy Each
722.00
M34153 MitoProbe™ Transition Pore Assay Kit, for flow cytometry Each
513.65
Online Exclusive
684.00
Save 170.35 (25%)
  1. Nicholls, DG (2004) Mitochondrial Membrane Potential and Aging. Aging Cell 3: 35-40.
  2. Tirmenstein, MA, Hu, CX, Gales TL, et al. (2002) Effects of Troglitazone on HepG2 Viability and Mitochondrial Function Toxicol. Sci.69: 131-8.
  3. O'Brien PJ, Irwin W, Diaz D, et al. (2006) High Concordance of Drug-Induced Human Hepatotoxicity With in Vitro Cytotoxicity Measured in a Novel Cell-Based Model Using High Content Screening. Arch Toxicol 80: 580-604.
  4. Dykens JA, Will Y. (2007) The Significance of Mitochondrial Toxicity Testing in Drug Development. Drug Discovery Today 12:777-85.
  5. Dykens JA, Jamieson JD, Marroquin LD, et al. (2008) In Vitro Assessment of Mitochondrial Dysfunction and Cytotoxicity of Nefazodone, Trazodone, and Buspirone. Toxicol Sci 103: 335-45.
  6. Abraham VC, Towne DL, Waring JF, et al. (2008) Application of a High-Content Multiparameter Cytotoxicity Assay to Prioritize Compounds Based on Toxicity Potential in Humans. J Biomol Screen 13: 527-37.
  7. He L, He T, Farrar S et al. (2017) Antioxidants Maintain Cellular Redox Homeostasis by Elimination of Reactive Oxygen Species. Cell Physiol Biochem 44: 532-553.
  8. Giorgi C, Romagnoli A, Pinton P et al. (2008) Ca2+ Signaling, Mitochondria and Cell Death. Curr Mol Med 8: 119-130.
  9.  
Related categories
decorative image
decorative image
decorative image
decorative image
Tools and handbooks
Screen-Shot-2024-02-08-at-13.29.24
Fluorescence SpectraViewer
Supports all levels of experimental complexity, use the tool to compare excitation and emission spectra of fluorophores and reagents.
Access tool
decorative image
Stain-iT Cell Staining Simulator
Visualize staining your cell without wasting your reagents, antibodies, or time.
Try now
Learn how
 decorative image
Flow cytometry protocols handbook
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
Request the handbook
decorative image
Imaging protocol handbook
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies. 
Request the handbook
Support
Cell Analysis Support Center
Find technical information, tips and tricks, and answers to everyday problems.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments