TrypLE reagent product image
Gibco TrypLE gentle cell dissociation reagents
Ideal for both serum-supplemented and serum-free conditions, TrypLE enzymes are an answer to animal origin-free trypsin-like enzymes. Exhibiting a similar pH activity profile to trypsin, this enzyme cleaves at arginine and lysine. With the TrypLE enzyme’s animal-free origin, hazards from potential pathogenic contaminants have been eliminated; and subject to a controlled fermentation process, TrypLE enzymes are available for supply at any scale.
The TrypLE enzyme is designed to seamlessly fit your workflow. TrypLE enzymes can be substituted for trypsin in existing protocols, is room temperature stable and, in addition, is exceedingly gentle on cells. So gentle, in fact, that the TrypLE enzyme requires no inhibiting agent. Instead, it is inactivated by dilution alone. In addition, TrypLE enzymes boast similar dissociation kinetics to porcine trypsin as well as comparable cell replating, proliferation kinetics, and long-term maintenance.
TrypLE products offer a greener, more economical cell dissociation. For more information, see our TrypLE enzyme resources
Choose the right TrypLE enzymes for your cell culture
 TrypLE ExpressTrypLE Select
CharacteristicsGentler, stabler, and cost comparableGentler, stabler, animal and human-component free
Phenol redAvailable with or without phenol redPhenol red-free
Use caseGeneral purposeBioproduction/industrial application
Origin sourceFungus
(animal origin-free)
Fungus
(animal origin-free)
Manufacturing standardscGMP manufacturingcGMP, manufactured on dedicated animal origin-free machinery
Storage temperatureRoom temperatureRoom temperature
Higher cell viability
Inactivation methodInhibition by dilutionInhibition by dilution
A clear bottle on white background containing TrypLE Express Enzyme, with Phenol red
TrypLE Express reagents
Using the same recombinant fungal trypsin-like protease, the TrypLE Express reagent is manufactured to accommodate pricing comparable to standard trypsin. Unlike TrypLE Select formulations, TrypLE Express reagents come with and without phenol red.
For data on release times, stability statistics, and purity and plating efficiency, please see TrypLE Express performance data below.
Catalog # Name Size Price (USD) Qty
12605010 TrypLE™ Express Enzyme (1X), phenol red, 100 mL Each
37.65
Online Exclusive
42.21
Save 4.56 (11%)
12605093 TrypLE™ Express Enzyme (1X), phenol red, 5 L Each
923.65
Online Exclusive
1,032.00
Save 108.35 (10%)
12604021 TrypLE™ Express Enzyme (1X), no phenol red, 500 mL Each
112.65
Online Exclusive
123.00
Save 10.35 (8%)
12604054 TrypLE™ Express Enzyme (1X), no phenol red, 5 L Each
933.65
Online Exclusive
1,028.00
Save 94.35 (9%)
12605028 TrypLE™ Express Enzyme (1X), phenol red, 500 mL Each
109.65
Online Exclusive
123.00
Save 13.35 (11%)
12604013 TrypLE™ Express Enzyme (1X), no phenol red, 100 mL Each
38.65
Online Exclusive
42.22
Save 3.57 (8%)
12605036 TrypLE™ Express Enzyme (1X), phenol red, 20 x 100mL Each
592.65
Online Exclusive
662.00
Save 69.35 (10%)
12604039 TrypLE™ Express Enzyme (1X), no phenol red, 20 x 100mL Each
593.65
Online Exclusive
654.00
Save 60.35 (9%)
TrypLE Express performance data
Cell line release times and viability
Triplicate cultures of each cell line, serially passed six times over, were centrifugally washed, counted, and subcultured post-enzyme-dissociation. The following values represent replicate averages over these six passages.
Cell lineMediaMean time required
for cell release
Mean viability
VEROVP-SFM5 min100%
VEROMEM + 5% FBS8 min100%
VEROOptiPro7 min98%
PK-15MEM + 10% FBS27 min98%
PK-15OptiPro11 min99%
MDCKOptiPro28 min98%
MDBKDMEM + 5% FBS15 min100%
A549DMEM + 10% FBS9 min98%
293FDMEM + 5% FBS2 min97%
TrypLE Express reagent outperforms trypsin in preserving cell-surface epitope expression
Histogram showing how TrypLE Express reagent preserves CD2-APC better than trypsin
Figure 1. TrypLE Express reagent preserves cell-surface epitope expression. Jurkat cells were treated with 0.05% trypsin (A) or TrypLE Express reagent (B) for a period of up to 20 minutes. Cell-surface CD2 levels were then measured via flow cytometry with an APC-conjugated anti-CD2 monoclonal antibody. As demonstrated, cells treated with 0.05% trypsin show a clear time-dependent reduction in CD2 levels, those treated with TrypLE Express reagent exhibit no measurable CD2 loss.
Flow cytometry analysis: cell dissociation from mouse CNS tissue
Bar chart of viability vs annexin V when using TrypLE enzymes or Accutase
Figure 2. Cell viability is better with TrypLE reagent than with Accutase reagent​.
Dissociation time: 10 min; Viability (%) +/- SEM​. Adapted from data shown in Panchision DM, et al. Stem Cells, 25:1560 (2007). doi:10.1634/stemcells.2006-0260.
Bar chart of percent CD24-positive cells for both TrypLE reagent and papain treatments
Figure 3. The CD24 epitope is retained with TrypLE whereas use of papain illustrates complete loss of the CD24 epitope. Adapted from data shown in Panchision DM, et al. Stem Cells, 25:1560 (2007). doi:10.1634/stemcells.2006-0260.
Workflow benefits when using TrypLE enzymes
TrypLE enzymes simplify your workflow. For example, cells, treated with either animal trypsin or TrypLE Express, and replated with OptiPro SFM were left unwashed post-dissociation. In addition, no protease inhibitors were used. 24 hours later, morphology is recorded. As seen below, TrypLE enzyme-treated cells retain their structure, while trypsin cells experience further cellular degradation. 
Animal trypsin
Brightfield image of cultured cells after trypsin treatment
TrypLE Express
Brightfield image of cultured cells after TrypLE enzyme treatment
Figure 4. TrypLE enzyme-treated cells retain their structure when left unwashed after dissociation without protease inhibitors. While TrypLE enzyme-treated cells maintained their morphology under these conditions, trypsin-treated cells experienced further degradation.
Cell line release times and viability
Triplicate cultures of each cell line, serially passed six times over, were centrifugally washed, counted, and subcultured post-enzyme-dissociation. The following values represent replicate averages over these six passages.
Cell lineMediaMean time required
for cell release
Mean viability
VEROVP-SFM5 min100%
VEROMEM + 5% FBS8 min100%
VEROOptiPro7 min98%
PK-15MEM + 10% FBS27 min98%
PK-15OptiPro11 min99%
MDCKOptiPro28 min98%
MDBKDMEM + 5% FBS15 min100%
A549DMEM + 10% FBS9 min98%
293FDMEM + 5% FBS2 min97%
TrypLE Express reagent outperforms trypsin in preserving cell-surface epitope expression
Histogram showing how TrypLE Express reagent preserves CD2-APC better than trypsin
Figure 1. TrypLE Express reagent preserves cell-surface epitope expression. Jurkat cells were treated with 0.05% trypsin (A) or TrypLE Express reagent (B) for a period of up to 20 minutes. Cell-surface CD2 levels were then measured via flow cytometry with an APC-conjugated anti-CD2 monoclonal antibody. As demonstrated, cells treated with 0.05% trypsin show a clear time-dependent reduction in CD2 levels, those treated with TrypLE Express reagent exhibit no measurable CD2 loss.
Flow cytometry analysis: cell dissociation from mouse CNS tissue
Bar chart of viability vs annexin V when using TrypLE enzymes or Accutase
Figure 2. Cell viability is better with TrypLE reagent than with Accutase reagent​.
Dissociation time: 10 min; Viability (%) +/- SEM​. Adapted from data shown in Panchision DM, et al. Stem Cells, 25:1560 (2007). doi:10.1634/stemcells.2006-0260.
Bar chart of percent CD24-positive cells for both TrypLE reagent and papain treatments
Figure 3. The CD24 epitope is retained with TrypLE whereas use of papain illustrates complete loss of the CD24 epitope. Adapted from data shown in Panchision DM, et al. Stem Cells, 25:1560 (2007). doi:10.1634/stemcells.2006-0260.
Workflow benefits when using TrypLE enzymes
TrypLE enzymes simplify your workflow. For example, cells, treated with either animal trypsin or TrypLE Express, and replated with OptiPro SFM were left unwashed post-dissociation. In addition, no protease inhibitors were used. 24 hours later, morphology is recorded. As seen below, TrypLE enzyme-treated cells retain their structure, while trypsin cells experience further cellular degradation. 
Animal trypsin
Brightfield image of cultured cells after trypsin treatment
TrypLE Express
Brightfield image of cultured cells after TrypLE enzyme treatment
Figure 4. TrypLE enzyme-treated cells retain their structure when left unwashed after dissociation without protease inhibitors. While TrypLE enzyme-treated cells maintained their morphology under these conditions, trypsin-treated cells experienced further degradation.
A clear bottle with a white label on a white background containing TrypLE Select Enzyme (1X), Phenol red free
TrypLE Select reagents
TrypLE Select enzymes are manufactured on dedicated animal origin-free machinery and designed specifically for the bioproduction industry. Select enzymes come in two concentrations, 1X and 10X, and are phenol red-free. Using 10X at full strength can rapidly, but gently, dissociate strongly adherent cells.
For data on release times, stability statistics, and purity and plating efficiency, please see TrypLE Select data below.
What's new: Gibco TrypLE Select reagent in 5 and 10 L Aegis 5–14 film BPCs.
Easily scale up with the new 5 L and 10 L TrypLE Select catalog BPCs. The formulation of TrypLE Select reagent has not changed, so you will get the same level of performance you have come to rely on, including:
  • Animal origin-free formulation that can reduce the risk of contamination. 
  • Recombinant technology to help ensure high purity and consistency from batch to batch.
  • Gentler on cells compared to trypsin, leading to higher viability and less damage during the dissociation process. 
  • Activity and stability over a wide range of temperatures and storage conditions.
TrypLE Select performance data
Cell release times
The following cell lines were treated with TrypLE Select enzymes at either 1X or 10X concentrations. Their mean cell release times and mean viabilities were observed and recorded.
 (1x)(10x)
StrengthCell linesMediaMean cell
release time
Mean
viability
Mean cell
release time
Mean
viability
Adherent293FDMEM + 5% FBS2 min97%  
VEROVP-SFM5 min100%  
VEROEMEM + 5% FBS8 min100%  
VEROOptiPRO SFM7 min98%  
CHO-K1CHO III A7 min95%  
Strongly adherentPK-15EMEM + 10% FBS27 min98%11.7±2.0 min99.4%±0.3%
PK-15Gibco OptiPRO SFM11 min99%  
MDCKOptiPRO SFM28 min98%15.3±0.8 min98.7%±0.5%
MDCKDMEM + 5% FBS15 min100%8.0±1.3 min98.8%±0.5%
A549DMEM + 10% FBS9 min98%5.2±0.3 min99.1%±0.5%
MSCDMEM + 10% MSC FBS  7.8±1.2 min98.7%±1.3%
MSCGibco StemPro MSC SFM  8.7±1.5 min97.5%±0.1%
Stability statistics
At various temperatures, the TrypLE Select enzyme shows superior stability compared to either crude or purified porcine trypsin. Even after eight weeks at 37°C, the TrypLE Select enzyme maintained 50% activity while, after only two days, purified porcine trypsin enzyme activity rapidly decreased. 
Bar chart of TrypLE Select enzyme activity over 8 weeks
Figure 5. TrypLE Select enzyme stability. TrypLE Select enzyme shows superior stability compared to crude or purified porcine trypsin at various storage temperatures. Even after eight weeks at 37°C, 50% enzyme activity was still seen. All samples were stored in the dark.
Bar chart of porcine trypsin activity over 8 weeks
Figure 6. Purified porcine trypsin stability. Enzyme activity decreased rapidly in samples of purified porcine trypsin stored at 37°C.
Plating efficiency
Cells detached with TrypLE Select enzymes at 100–200 cells per well, have higher plating efficiency than those treated with porcine trypsin. 
Bar chart of percent animal enzyme control for 2 cell lines and TrypLE Select enzyme and animal trypsin treatments
Figure 7. TrypLE Select enzymes increases plating efficiency. Cells detached with TrypLE Select enzymes exhibit higher plating efficiency than cells detached with porcine trypsin.
Cell release times
The following cell lines were treated with TrypLE Select enzymes at either 1X or 10X concentrations. Their mean cell release times and mean viabilities were observed and recorded.
 (1x)(10x)
StrengthCell linesMediaMean cell
release time
Mean
viability
Mean cell
release time
Mean
viability
Adherent293FDMEM + 5% FBS2 min97%  
VEROVP-SFM5 min100%  
VEROEMEM + 5% FBS8 min100%  
VEROOptiPRO SFM7 min98%  
CHO-K1CHO III A7 min95%  
Strongly adherentPK-15EMEM + 10% FBS27 min98%11.7±2.0 min99.4%±0.3%
PK-15Gibco OptiPRO SFM11 min99%  
MDCKOptiPRO SFM28 min98%15.3±0.8 min98.7%±0.5%
MDCKDMEM + 5% FBS15 min100%8.0±1.3 min98.8%±0.5%
A549DMEM + 10% FBS9 min98%5.2±0.3 min99.1%±0.5%
MSCDMEM + 10% MSC FBS  7.8±1.2 min98.7%±1.3%
MSCGibco StemPro MSC SFM  8.7±1.5 min97.5%±0.1%
Stability statistics
At various temperatures, the TrypLE Select enzyme shows superior stability compared to either crude or purified porcine trypsin. Even after eight weeks at 37°C, the TrypLE Select enzyme maintained 50% activity while, after only two days, purified porcine trypsin enzyme activity rapidly decreased. 
Bar chart of TrypLE Select enzyme activity over 8 weeks
Figure 5. TrypLE Select enzyme stability. TrypLE Select enzyme shows superior stability compared to crude or purified porcine trypsin at various storage temperatures. Even after eight weeks at 37°C, 50% enzyme activity was still seen. All samples were stored in the dark.
Bar chart of porcine trypsin activity over 8 weeks
Figure 6. Purified porcine trypsin stability. Enzyme activity decreased rapidly in samples of purified porcine trypsin stored at 37°C.
Plating efficiency
Cells detached with TrypLE Select enzymes at 100–200 cells per well, have higher plating efficiency than those treated with porcine trypsin. 
Bar chart of percent animal enzyme control for 2 cell lines and TrypLE Select enzyme and animal trypsin treatments
Figure 7. TrypLE Select enzymes increases plating efficiency. Cells detached with TrypLE Select enzymes exhibit higher plating efficiency than cells detached with porcine trypsin.
TrypLE enzyme resources
Try the Gibco MediaSelect Tool
Easily find a media solution suited to your specific application. Use this interactive tool to select classical basal, protein expression, or viral vector media.
Your selections will allow you to choose from a catalog menu of media – or customize your own formulation to fit your experimental setup and budget. Where available, you can compare media formulations side-by-side.
Find your media with the Gibco MediaSelect Tool
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Gibco Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments