AAV-MAX viral particle
Adeno-Associated Viral Vector Production
Cost-effective, scalable adeno-associated virus (AAV) vector production is important to meet commercial demand, and a smooth scale-up to clinical production is essential. To address these needs, we have created a complete AAV production system to help reduce production costs and streamline your transition from research to clinical scale.
Request information
The AAV-MAX system features:
  • High AAV titers—helps reduce production costs
  • Scalability—suspension system with scalable protocols from shake flask to bioreactor scale
  • Simplified workflow—streamlined protocol using helper virus-free triple transfection
  • Animal origin–free (AOF)
  • Clonal 293F-derived producer cells
Figure 1. Components of the Gibco AAV-MAX Helper Free AAV Production System.
The Gibco AAV-MAX Helper Free AAV Production System includes clonal HEK293F–derived suspension cells, chemically defined, animal origin-free, serum-free media, a proprietary transfection reagent, a booster, a novel production enhancer, and a polysorbate 20-based lysis buffer.
Multi-component system optimized for improved AAV production
The AAV-MAX system provides a scalable and high-titer a platform for suspension AAV production. It is based on a high-density suspension culture of clonal 293F–derived Viral Production Cells 2.0 in Viral Production Medium. Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel production enhancer (Figure 1).
Optimized and fully integrated system
The components of the AAV-MAX system are designed to work synergistically, resulting in maximal titers and eliminating the need to optimize reagents and protocols. The complete system produces higher titers compared to swapping out individual components and using conventional polyethyleneimine (PEI)-based transfection reagents (Figure 2).
Figure 2. The complete system delivers maximum AAV titers compared to alternative transfection reagents. AAV2 and AAV6 were produced at a 30 mL scale in 125 mL shake flasks using the AAV-MAX system. These data highlight that the highest titers were achieved using the complete system compared to the reduced titers observed when using other transfection reagents. Titers were measured by qPCR. The data was normalized to the titer (vg/mL) of the complete AAV-MAX system.
High titers across multiple serotypes
The innovative AAV-MAX Helper-Free AAV Production System is a complete optimized suspension system that allows you to seamlessly and efficiently produce your AAV vector with high titers across multiple AAV serotypes.
Figure 3. The system yields high titers across multiple AAV serotypes. The AAV-MAX system was used to produce 5 AAV serotypes at a 30 mL scale in 125 mL shake flasks. These data show that high genome titers, high cell viabilities, and high viable cell densities (VCD) were achieved for each of the 5 serotypes. Titers were measured by qPCR.
A cost-effective AAV production solution
The AAV-MAX system can help you obtain the same viral titer in less volume compared to PEI-based production systems, which translates to savings in media, downstream purification, lab space, and labor.
This advantage can mitigate contamination risks by working with smaller batch sizes. In addition, the AAV-MAX System requires less plasmid DNA per 1x106 cells than alternative transfection reagents, which further drives production costs down.
Switch to the AAV-MAX system and save
  • Save 50% on average compared to alternative PEI-based suspension systems
  • Cut plasmid DNA cost by 25%
Scaling to meet demand
Challenges associated with existing AAV production systems include low titers, high cost of cGMP plasmid DNA, low scalability, and lack of fit for purpose, cGMP reagents. The AAV-MAX system can help overcome these challenges by allowing you to produce high AAV titers using pre-optimized and regulatory-compliant* reagents that were developed for AAV production in a scalable suspension platform where volumetric viral titers are maintained as you scale up.
Ordering information
Catalog # Name Size Price (USD) Qty
A51218 Viral Production Cells 2.0, 6 vials Each
6,350.00
A50515 AAV-MAX Transfection Kit, 1 kit for 1 L of production Each
1,085.27
Special offer
Online exclusive
1,172.00
Save 86.73 (7%)
A50516 AAV-MAX Transfection Kit, 1 kit for 10 L of production Each
8,996.50
Special offer
Online exclusive
9,470.00
Save 473.50 (5%)
A4817901 Viral Production Medium, 1 L Each
253.80
Special offer
Online exclusive
282.00
Save 28.20 (10%)
A51217 AAV-MAX Helper-Free AAV Production System Kit Each
2,080.00
A4983901 Viral-Plex™ Complexation Buffer Each
49.89
Special offer
Online exclusive
54.50
Save 4.61 (8%)
A50520 AAV-MAX Lysis Buffer Each
167.40
Special offer
Online exclusive
186.00
Save 18.60 (10%)
A4817902 Viral Production Medium, 6 L Each
1,376.40
Special offer
Online exclusive
1,480.00
Save 103.60 (7%)
A4817903 Viral Production Medium, 10 L Each
2,134.65
Online Exclusive
2,280.00
Save 145.35 (6%)
A49784 Viral Production Cells 2.0, 1 vial Each
1,516.00
Looking for a partner to help accelerate your discovery?
Our dedicated staff of R&D scientists and project managers are ready to support you with end-to-end services for every step in your Adeno-associated virus (AAV) RUO production. Learn more about our viral vector production services.
Resources
Support
* Available with CTS AAV-MAX Helper Free Production System, set for release in 2022.