Gibco brand media provide the broadest selection of specialty products that are optimized for neural stem cell populations, many of which are free of animal derived components. These media are designed to help minimize adaptation time, maximize cell performance, and meet regulatory requirements.
Media
  • Neurobasal Plus Medium: an optimized basal medium formulation that enables 50% higher survival rate of neural cells when compared to classic Neurobasal medium and B-27 Supplement.
  • Neurobasal-A Medium: specially formulated basal medium designed to help meet the special requirements of neuronal cells in culture.
  • CTS Neurobasal Medium: a serum-free basal medium for the long-term viability of prenatal and embryonic hippocampal and other neurons of the CNS
  • CTS Neurobasal -A Medium: a serum-free basal medium for the long-term viability of postnatal and adult hippocampal and other neurons of the CN
  • StemPro NSC SFM: Specifically formulated for serum-free growth and expansion of human neural stem cells
Serum replacement
  • CTS KnockOut SR XenoFree Medium: enables the growth and expansion of hESCs and hiPSCs in a cell culture medium containing only human-derived or human recombinant proteins, to help facilitate the transition of hESC and hiPSC research from the bench to the clinic
  • Knockout Serum Replacement: serum-free formulation designed to replace FBS in neural stem cell culture
Supplements
  • B-27 Plus Supplement: a serum-free supplement that provides the highest rate of in vitro survival of cultured neurons when used with Neurobasal Plus Medium
  • CultureOne Supplement: a chemically-defined, serum-free supplement designed to help improve the differentiation of neural stem cells (NSCs) to neurons
  • CTS B-27 Supplement, XenoFree: can be used to support induction of human neural stem cells (hNSCs) from pluripotent stem cells (PSCs), expansion of hNSCs, differentiation of hNSCs, and maintenance of mature differentiated neurons in culture
  • CTS N-2 Supplement: a serum-free supplement for the growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype
  • StemPro Neural Supplement: serum-free supplement for growth and expansion of human and rat neural stem cells (NSCs) and progenitor cells (PCs)
  • N-2 Supplement: serum substitute for embryonic neurons/CNS progenitor cells
  • G-5 Supplement: serum-free supplement for the growth and expression of glial cells (normal and tumor) of astrocytic phenotype
  • Gibco growth factors: growth factors are cell signaling proteins that play important roles in cell proliferation, differentiation, and survival
Matrices and substrates
  • CELLstart: the first xeno-free substrate for attachment and expansion of human embryonic, mesenchymal, and neural stem cells that is fully-defined, has better lot-to-lot consistency and is produced under cGMP
For Research Use Only. Not for use in diagnostic procedures.