NuPAGE Bis-Tris Gels and Bolt Bis-Tris Plus Gels are high-performance precast polyacrylamide gels developed to provide optimal separation of a wide range of protein sizes under denaturing conditions. The neutral pH of the Bis-Tris chemistry helps minimize protein modifications and reduce protein degradation. As a result, NuPAGE and Bolt gels are a good choice for any application where protein integrity is crucial.
NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gel specifications
NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gels offer:
  • Excellent protein integrity—neutral pH helps reduce protein degradation
  • Wide range of molecular weight separation—select the right gel and running buffer to achieve the optimal separation of your proteins
  • Fast run times—separate your proteins in 20–35 minutes
  • Long shelf life—up to 16 months at room temperature
  • High-capacity wells—WedgeWell format wells are wedge-shaped wells that double the well sample loading capacity and make it easy to load your sample (no specialized pipette tips required)
 NuPAGE Bis-TrisBolt Bis-Tris Plus
Select forHigher sample throughput, increased sample volume capacityFaster run times, increased sample volume capacity
Available gel sizesMini: 8 cm x 8 cm (1.0 or 1.5 mm thick)
Midi: 8 cm x 13 cm (1.0 mm thick)
Mini: 8 cm x 8 cm (1.0 mm thick)
Available Wells configurations*WedgeWell format Midi: 12+2, 20, 26 wells
Mini: 1, 9, 10, 12, 15, 17, 2D-well, IPG wells
Midi: 12+2, 20, 26 wells
WedgeWell format Mini: 10, 12, 15, 17 well
Storage conditions4–25°C
Shelf lifeup to 16 months**16 months
High lot-to-lot consistencyCoefficient of variation (CV) of only 2% for Rf values (migration)Coefficient of variation (CV) of only 2% for Rf values (migration)
Recommended sample bufferNuPAGE LDS Sample BufferBolt LDS sample buffer
Recommended running buffersNuPAGE MES SDS or NuPAGE MOPS SDS Running BufferBolt MES SDS or Bolt MOPS SDS running buffer
Run time35 min (MES buffer)
50 min (MOPS Buffer)
20 min (MES buffer)
35 min (MOPS buffer)
Recommended transfer bufferNuPAGE Transfer BufferBolt Transfer Buffer
Gel chemistryBis-TrisBis-Tris Plus
Available polyacrylamide concentrations8%, 10%, 12%, 4–12%
Separation range15 kDA to 260 kDa (MOPS buffer)
3.5 kDA to 160 kDa (MES buffer)
For use with (equipment) mini gelsMini Gel Tank or XCell SureLock Mini-Cell
For use with (equipment) midi gelsSureLock Tandem Midi Gel Tank, Invitrogen XCell4 SureLock Midi-Cell or Bio-Rad Criterion (with adapters only)Not available in midi size
Mode of separationMolecular weight
ApplicationSDS-PAGE
*Not all percentages are available in every well type
**Shelf life varies depending on gel format
Benefits of the Bis-Tris and Bis-Tris plus gel chemistry
NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, we recommend using buffers containing SDS. The gels can be run using MES SDS running buffer to better resolve smaller proteins (1–200 kDa) and MOPS SDS running buffer to resolve medium- to large-size proteins (14–260 kDa).
Bis-Tris Gels use a Bis-Tris discontinuous buffer system involving three ions:
  • Chloride (–) is from the gel buffer and serves as a leading ion due to its high affinity to the anode relative to other anions in the system. The gel buffer ions are bis-tris (+) and Cl (–) (pH 6.4).
  • MES or MOPS (–) serves as the trailing ion in the running buffer. The running buffer ions are tris (+), MOPS/MES (–) and dodecylsulfate (pH 7.3–7.7).
  • Bis-Tris (+) is the common ion present in the gel buffer and running buffer. The combination of lower pH gel buffer (pH 6.4) and the running buffer (pH 7.3–7.7) results in a significantly lower operating pH of 7 during electrophoresis
Reduced protein degradation and sharp, straight bands
Optimized performance of NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gels
Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gels are Bis-Tris HCl buffered (pH 6.4) and have an operating pH of about 7.0. This neutral pH paired with a unique, gentle sample preparation protocol means your protein samples always reside in mild, nonacidic conditions, preserving the integrity of the proteins and minimizing protein modifications (see next section). This helps to ensure highly sensitive, accurate western blot results every time.
Western blot analysis using a Bolt Bis-Tris Plus gel compared to tris-glycine gels
Figure 1. Greater sensitivity with Bolt Bis-Tris Plus gels. Total cell extracts from A431 cells were transferred to NC and PVDF membranes from a 4–12% Bolt Bis-Tris Plus gel, and 4–20% Tris-glycine precast gel using the iBlot 2 Gel Transfer Device. The cells were treated with 100 ng/mL of human epidermal growth factor (hEGF) to up-regulate expression of the phospho-EGF receptor. The protein loads of the cell extracts ranged from 20 μg to 1.2 μg of extract. The blots were processed on the iBind Western System using a 1:200 dilution of Phospho-EGF Receptor (Tyr1068) (1H12) Mouse mAb (Cell Signaling Technology) and a 1:2,000 dilution of anti-mouse HRP secondary antibody.
Excellent sample integrity
Sample preparation is a crucial step in successful protein analysis. The pH of the traditional Laemmli-style sample buffer changes from 6.8 to 5.2 when heated at 100°C. This lower pH is known to induce aspartyl-prolyl (Asp-Pro) peptide bond cleavage, which leads to protein degradation. By maintaining a >7.0 pH environment, the Invitrogen NuPAGE LDS Sample Buffer and Bolt LDS Sample Buffer preserve protein integrity by minimizing this Asp-Pro cleavage. As seen below, the sample integrity is maintained throughout electrophoresis with the Invitrogen NuPAGE system; protein degradation is seen in samples prepared with Laemmli (Tris-glycine) sample buffer. The Invitrogen NuPAGE Antioxidant running buffer additive greatly minimizes protein oxidation during electrophoresis and keeps reduced protein bands sharp and clear.
Figure 2. High sample integrity. Integrity of samples is maintained throughout electrophoresis with the NuPAGE Bis-Tris Gel System (left), compared to samples prepared with Laemmli (tris-glycine) sample buffer (right).
Sharp, straight bands
Neutral-pH buffers in NuPAGE Bis-Tris and Bolt Bis-Tris Plus Gels deliver sharp straight bands. During separation, the gels operate close to pH 7. In the Laemmli system (Tris-glycine), the gel is run at basic pH (pH ~9.5). At high pH the residual unpolymerized acrylamide can react with cysteine and lysine residues of the proteins being separated. This may affect downstream analysis such as western blot transfer. At the neutral pH of NuPAGE Bis-Tris and Bolt Bis-Tris Gels, these reactions take place several orders of magnitude more slowly than at the basic pH of Tris-glycine gels, resulting in sharper bands and better resolution.
NuPAGE Bis-Tris gels provide sharp straight bands compared to competitor tris glycine gels
Figure 3. Protein separation comparing NuPAGE Bis-Tris Gel and traditional tris-glycine gel. The samples listed below were run on (A) a NuPAGE 4–12% Bis-Tris Gel in Invitrogen NuPAGE MES SDS Running Buffer or (B) another manufacturer’s 4–20% tris-glycine gel. Note the high resolution of the sample bands in the NuPAGE protein gel. The poorly resolved “fuzzy” bands seen in the other manufacturer’s gel are a result of reoxidation of some disulfide bonds within the sample, leading to slight changes in migration rates. Lanes 1, 6, 7, 10: Invitrogen Mark12 Unstained Standard; lane 2: high protein load (4 proteins, 6 µg/protein); lane 3: Broad Range Standard; lanes 4, 5: E. coli extract; lane 8: E-PAGE SeeBlue Pre-stained Standard; lane 9: Invitrogen MultiMark Multi-Colored Protein Standard.
Band integrity
NuPAGE and Bolt gels preserve protein integrity with neutral-pH gel chemistry. This formulation minimizes protein modifications during electrophoresis.
Figure 4. A western blot of a Bolt Bis-Tris Plus gel shows clean, sharp protein signals corresponding to only full-length proteins, whereas a western blot of a Bio-Rad TGX gel shows multiple low molecular weight degradation products. Protein kinases implicated in cancer (IKKϐ, EPHB3, HCK, MAPK14, FLT1 and DDR2) were analyzed on a Bolt Bis-Tris Plus Gel and a Bio-Rad TGX tris-glycine gel. The purified kinases (50 ng each), along with MagicMark protein standard and purified recombinant GST protein, were loaded on a 10-well, 4–12% Bolt gel and a 10-well, 4–20% Bio-Rad TGX gel. The samples were separated and transferred to 0.45-μm PVDF membranes using the respective manufacturers’ protocols. Immunodetection was performed using an anti-GST antibody and WesternBreeze chemiluminescence detection.
Band resolution
NuPAGE and Bolt Bis-Tris Plus Gels have better resolving power than Bio-Rad’s gels. With 10% greater resolving distance, optimized gel chemistry, and gradient format, you can see more protein bands resolved on a Bolt gel. In experimental analysis, you can see two separate bands for the insulin B chain and insulin A chain, which have similar molecular weights. The protein bands on the Bolt gel appear straight and the signal intensities are even across the entire bands, whereas Bio-Rad’s gel show fewer (Figure 5C), or poorly resolved (Figure 5B, lane 2, 7 and 8) protein bands.
Figure 5. Bolt Bis-Tris Gel band resolution—comparison of Bio-Rad's stained protein gels. (A) Bolt Bis-Tris gradient gel stained with Invitrogen SimplyBlue SafeStain. (B) Bio-Rad's tris-glycine gradient gel stained with competitor’s stain. (C) Enlargement of a section of lane 5 from the two gels shows the insulin B chain (3.5 kDa) and insulin A chain (2.5 kDa).
Publication-quality protein electrophoresis data is easily generated using the higher-throughput NuPAGE WedgeWell format Bis-Tris midi gels and the SureLock Tandem Midi Gel Tank. The gel produces crisp bands and straight protein lanes.
Figure 6. High sample volume capacity of Bolt Bis-Tris Plus gels. (A) Increasing volumes (40–70μL) of a fluorescent protein standard were loaded in every other lane of an Invitrogen Bolt 4–12% Bis-Tris Plus 10-well Gel. (B) Increasing volumes (10–40 μL) of the same protein standard were loaded in every other lane of Bio-Rad's 4–20% 10-well gel. Sample spillover and cross-well contamination is observed as signals from the sample loaded in the adjacent wells.
NuPAGE Midi gels with high loading capacity wells
WedgeWell format wells are wedge-shaped wells that double the sample loading capacity of traditional wells. WedgeWell format Midi protein gels allow the separation of up to 26 protein samples in the same run, without compromising on sample loading volume. Here are the key benefits of WedgeWell Midi gels:
  • Results you can trust—rigorously tested to help ensure reliable, reproducible performance, lot after lot
  • Easy sample loading—reduced sample spill-over and cross-well contamination
  • Load more, see more—easier detection of dilute samples and low abundance proteins
  • More data in less time—obtain the information you need, faster
  • Reduced cost—lower cost per sample and reduced running buffer required per sample
  • Faster run times—up to 10 min faster than conventional gels
NuPAGE Bis-Tris Midi WedgeWellformat gels meet the performance, reliability, and consistency standards Invitrogen precast gels are known for.
See our Protein Gel Performance Guarantee
Recommended sample loading volumes for WedgeWell format wells
Well sizeRecommended loading
volume (µL)
Maximum loading
volume (µL)
WedgeWell format midi gels
12 + 2 (large well)90100
12 + 2 (small well)3035
205060
263040
WedgeWell format mini gels
104060
123045
152035
171530
Midi gels
12 + 2 (large well)4560
12 + 2 (small well)1520
202530
261520
Mini gels*
91728
101525
121220
15915
17915
*1 mm thick gel
Excellent protein separation
NuPAGE Bis-Tris Midi WedgeWell format gels offer the same exceptional protein separation of conventional NuPAGE Bis-Tris Midi gels.
Coomassie stained protein gels
Figure 7. Straight, well-resolved bands for both WedgeWell and standard midi gels. Both gels were loaded as follows: Lanes 1, 20: 5 µL PageRuler Broad Range Unstained Protein Ladder (Cat. No. 26630); lanes 2–7: 10 µg, 8 µg, 6 µg, 4 µg, 2 µg, 1 µg HeLa lysate, respectively; lanes 8, 13: 5 µL Novex Mark12 Unstained Standard (Cat. No. LC5677); lanes 9–12: 240 ng, 180 ng, 120 ng, 60 ng of protein mix containing β-galactosidase, lactate dehydrogenase, and lysozyme; lanes 14–19: 10 µg, 8 µg, 6 µg, 4 µg, 2 µg, 1 µg E. coli lysate, respectively. Electrophoresis was conducted with NuPAGE MOPS running buffer. Gels were stained with SimplyBlue Safe Stain
Exceptional protein load capacity
NuPAGE Bis-Tris Midi WedgeWell format gels offer increase sample loading capacity without compromising on the performance of the separation.
Figure 8. Exceptional protein load capacity of NuPAGE Bis-Tris WedgeWell midi gels. Decreasing total amount (60–0.5 µg) of HEK293 cell lysate were denatured in NUPAGE SDS Sample Buffer, loaded in a NuPAGE Bis-Tris Wedgewell 4–12% 12+2 Well Midi Gel and run in the SureLock Tandem Midi Gel Tank. Equal amounts of HEK293 cell lysate were denatured in XT sample buffer, loaded in a Bio-Rad 4–12% XT12+2 well midi gel and run in the Criterion Cell Midi Gel Tank.
Comparison of electrophoresis and western blotting of cell lysates using NuPAGE midi gels versus Bio-Rad
The protein load capacities of Invitrogen NuPAGE Bis-Tris precast midi gels run in the SureLock Tandem Mid Gel Tank were compared against Bio-Rad Criterion midi gels run in a Bio-Rad Criterion Cell Midi Cell Tank using manufacturer instructions. Decreasing amounts of HEK 293 cell lysate prepared in RIPA lysis buffer (48–0.5 µg total protein) were denatured in the respective manufacturer’s sample buffer and subjected to electrophoresis using manufacturer instructions. The table below lists the samples, protein mass, and %RIPA buffer loaded in each lane.
Lane+1123456789101112+1
SampleSample bufferiBright Protein LadderHEK293 lysateiBright Protein LadderSample buffer
Load mass (µg)--484032241684210.5--
Load vol. (µL)332020202020202020202033
% RIPA buffer--4033.326.72013.36.73.31.70.80.4--
Invitrogen NuPAGE Bis-Tris gels outperformed Bio-Rad gels at higher lysate loads with blots from Bio-Rad gels showing band loss and smearing at higher loads for all targets investigated.
Comparison of protein blotting and western detection of cell lysates
NuPAGE gel system provides better sample integrity than the traditional Laemmli (tris-glycine) gel system
Figure 9: Western blots using NuPAGE 4-12% Bis-Tris midi gels generate sharper bands at greater protein and RIPA lysis buffer loads than Bio-Rad 4–20% TGX midi gels. A NuPAGE 4–12% Bis-Tris midi gel, 12+2 well, was loaded with decreasing total protein amount of HEK293 lysate, subjected to electrophoresis in a SureLock Tandem Midi Gel Tank and transferred onto a 0.45 µm PVDF membrane using the SureLock Tandem Blot Module and the Bio-Rad 4–20% TGX midi gel, 12+2 well, was subjected to electrophoresis in a Criterion Midi Cell Tank and transferred onto a 0.45 µm PVDF membrane using the Criterion Blotter. Both membranes were probed for three targets (vinculin, α-tubulin, and p23) using fluorescent immunodetection. Band loss and smearing can be seen at the higher loads for all targets on the Bio-Rad blot, while the blot for the NuPAGE Bis-Tris gel offers superior protein loading capacity above 24 µg and crisp, bright bands.
For immunodetection: membranes were blocked for 1 hour in 1X Blocker FL Fluorescent Blocking Buffer. For fluorescence: multiplex detection, the membranes were probed overnight with a mixture of primary antibodies diluted in blocking solution: Rabbit-anti Vinculin (1:15,000), Rat anti-α Tubulin (1:5,000)), and Mouse-anti p23 (1:5,000) followed by an incubation with secondary antibodies diluted at 1:5,000 in 1X Blocker FL: Donkey anti-Rabbit Alexa Fluor Plus 680, Donkey anti-Rat Alexa Fluor 488, and Donkey anti-Mouse Alexa Fluor Plus 800 for 1 hour. Membranes were imaged on an iBright Imaging System, with the same time and image adjustment settings for each blot.
Ordering Information
Catalog # Name Size Price (USD) Qty
WBT41212BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 12 + 2-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT41220BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 20-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT41226BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01012BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 12 + 2-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01020BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 20-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01026BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) 10 gels (1 box)
230.00
WG1201BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1202BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1203BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1401BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1402BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
206.65
Online exclusive
225.00
Save 18.35 (8%)
WG1403BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
207.65
Online exclusive
225.00
Save 17.35 (8%)
WG1001BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1002BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1003BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1201A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
222.65
Online exclusive
237.00
Save 14.35 (6%)
WG1202A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1203A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1401A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1402A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
224.65
Online exclusive
239.00
Save 14.35 (6%)
WG1403A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
216.65
Online exclusive
238.00
Save 21.35 (9%)
WG1001A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
238.00
WG1002A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
238.00
WG1003A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
237.00
Catalog # Name Size Price (USD) Qty
NW04120BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW04122BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
169.65
Online exclusive
181.00
Save 11.35 (6%)
NW04125BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
168.65
Online exclusive
181.00
Save 12.35 (7%)
NW04127BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
168.65
Online exclusive
181.00
Save 12.35 (7%)
NW00100BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00102BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00105BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
173.65
Online exclusive
181.00
Save 7.35 (4%)
NW00107BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00120BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00122BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00125BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00127BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00080BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
172.65
Online exclusive
181.00
Save 8.35 (5%)
NW00082BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
169.65
Online exclusive
181.00
Save 11.35 (6%)
NW00085BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00087BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
172.65
Online exclusive
181.00
Save 8.35 (5%)
Catalog # Name Size Price (USD) Qty
NP0321BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0322BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0323BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0329BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 17-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0327BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 9-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0335BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.5 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0336BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.5 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0324BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 1-well, 10 gels (1 box) Each
147.65
Online exclusive
164.00
Save 16.35 (10%)
NP0301BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0302BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0303BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0307BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 9-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0315BOX NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0316BOX NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0341BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0342BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
154.65
Online exclusive
164.00
Save 9.35 (6%)
NP0343BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
153.65
Online exclusive
164.00
Save 10.35 (6%)
NP0349BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 17-well, 10 Gels/Box Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NuPAGE Bis-Tris and Bolt Bis-Tris Plus Reagents
Catalog # Name Size Price (USD) Qty
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
22.34
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
176.00
NP0002 NuPAGE™ MES SDS Running Buffer (20X), 500 mL Each
102.65
Online exclusive
107.00
Save 4.35 (4%)
NP0060 NuPAGE™ MES SDS Buffer Kit (for Bis-Tris Gels) Each
128.65
Online exclusive
133.00
Save 4.35 (3%)
NP0001 NuPAGE™ MOPS SDS Running Buffer (20X), 500 mL Each
109.00
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
126.65
Online exclusive
132.00
Save 5.35 (4%)
NP0005 NuPAGE™ Antioxidant Each
39.33
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
18.65
Online exclusive
19.78
Save 1.13 (6%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
76.65
Online exclusive
80.75
Save 4.10 (5%)
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
33.80
NW2000 Mini Gel Tank and Blot Module Set Each
1,251.65
Online exclusive
1,302.00
Save 50.35 (4%)
A25977 Mini Gel Tank Each
700.65
Online exclusive
724.00
Save 23.35 (3%)
B1000 Mini Blot Module Each
457.00
Catalog # Name Size Price (USD) Qty
WBT41212BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 12 + 2-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT41220BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 20-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT41226BOX NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01012BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 12 + 2-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01020BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 20-well, 10 gels (1 box) 10 gels (1 box)
230.00
WBT01026BOX NuPAGE™ Bis-Tris Midi Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) 10 gels (1 box)
230.00
WG1201BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1202BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1203BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1401BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
212.65
Online exclusive
226.00
Save 13.35 (6%)
WG1402BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
206.65
Online exclusive
225.00
Save 18.35 (8%)
WG1403BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
207.65
Online exclusive
225.00
Save 17.35 (8%)
WG1001BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1002BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1003BOX NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, 10 gels (1 box) 10 gels (1 box)
225.00
WG1201A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
222.65
Online exclusive
237.00
Save 14.35 (6%)
WG1202A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1203A NuPAGE™ 10%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1401A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
223.65
Online exclusive
238.00
Save 14.35 (6%)
WG1402A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
224.65
Online exclusive
239.00
Save 14.35 (6%)
WG1403A NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
216.65
Online exclusive
238.00
Save 21.35 (9%)
WG1001A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 12+2-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
238.00
WG1002A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 20-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
238.00
WG1003A NuPAGE™ 8%, Bis-Tris, 1.0 mm, Midi Protein Gel, 26-well, w/adapters, 10 gels (1 box) 10 gels (1 box)
237.00
Catalog # Name Size Price (USD) Qty
NW04120BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW04122BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
169.65
Online exclusive
181.00
Save 11.35 (6%)
NW04125BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
168.65
Online exclusive
181.00
Save 12.35 (7%)
NW04127BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 4-12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
168.65
Online exclusive
181.00
Save 12.35 (7%)
NW00100BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00102BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00105BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
173.65
Online exclusive
181.00
Save 7.35 (4%)
NW00107BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00120BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00122BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00125BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00127BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 12%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
170.65
Online exclusive
181.00
Save 10.35 (6%)
NW00080BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
172.65
Online exclusive
181.00
Save 8.35 (5%)
NW00082BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
169.65
Online exclusive
181.00
Save 11.35 (6%)
NW00085BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
171.65
Online exclusive
181.00
Save 9.35 (5%)
NW00087BOX Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format, 10 gels (1 box) Each
172.65
Online exclusive
181.00
Save 8.35 (5%)
Catalog # Name Size Price (USD) Qty
NP0321BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0322BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0323BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0329BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 17-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0327BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 9-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0335BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.5 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0336BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.5 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0324BOX NuPAGE™ 4 to 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 1-well, 10 gels (1 box) Each
147.65
Online exclusive
164.00
Save 16.35 (10%)
NP0301BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0302BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0303BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0307BOX NuPAGE™ 10%, Bis-Tris, 1.0 mm, Mini Protein Gel, 9-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0315BOX NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 10-well, 10 gels (1 box) Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0316BOX NuPAGE™ 10%, Bis-Tris, 1.5 mm, Mini Protein Gel, 15-well, 10 gels (1 box) Each
159.65
Online exclusive
164.00
Save 4.35 (3%)
NP0341BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 10-well, 10 Gels/Box Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NP0342BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 12-well, 10 Gels/Box Each
154.65
Online exclusive
164.00
Save 9.35 (6%)
NP0343BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 15-well, 10 Gels/Box Each
153.65
Online exclusive
164.00
Save 10.35 (6%)
NP0349BOX NuPAGE™ 12%, Bis-Tris, 1.0 mm, Mini Protein Gel, 17-well, 10 Gels/Box Each
157.65
Online exclusive
164.00
Save 6.35 (4%)
NuPAGE Bis-Tris and Bolt Bis-Tris Plus Reagents
Catalog # Name Size Price (USD) Qty
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
22.34
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
176.00
NP0002 NuPAGE™ MES SDS Running Buffer (20X), 500 mL Each
102.65
Online exclusive
107.00
Save 4.35 (4%)
NP0060 NuPAGE™ MES SDS Buffer Kit (for Bis-Tris Gels) Each
128.65
Online exclusive
133.00
Save 4.35 (3%)
NP0001 NuPAGE™ MOPS SDS Running Buffer (20X), 500 mL Each
109.00
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
126.65
Online exclusive
132.00
Save 5.35 (4%)
NP0005 NuPAGE™ Antioxidant Each
39.33
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
18.65
Online exclusive
19.78
Save 1.13 (6%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
76.65
Online exclusive
80.75
Save 4.10 (5%)
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
33.80
NW2000 Mini Gel Tank and Blot Module Set Each
1,251.65
Online exclusive
1,302.00
Save 50.35 (4%)
A25977 Mini Gel Tank Each
700.65
Online exclusive
724.00
Save 23.35 (3%)
B1000 Mini Blot Module Each
457.00
Protein Gel Selection Guide
Protein Gel Selection Guide
Find an excellent mini or midi gel for your protein electrophoresis experiment.
Gel Selection Chart
Protein Gel Conversion Tool
Find an exceptional Invitrogen mini or midi gel to replace your current precast gel from another supplier.
Protein Gel Options Brochure
Gel Migration Chart
Download the Protein Gel Guide for a printable gel migration chart.
Still unsure which precast gel works best for your research needs? Download the Gel Selection Chart
Protein Gels Performance Guarantee
Protein Gels Performance Guarantee
We stand behind the performance of our protein gels.
Learn more
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.