Agarose I (Molecular Biology Grade)
Agarose I (Molecular Biology Grade)
Thermo Scientific™

Agarose I (Molecular Biology Grade)

Thermo Scientific Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lotRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
17850100 g
17852500 g
Catalog number 17850
Price (EUR)
369,00
Each
-
Add to cart
Quantity:
100 g
Price (EUR)
369,00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lot consistency for preparation of electrophoresis gels.

Features:

• A regular melting temperature agarose for electrophoresis requiring an exceptional level of purity with unequaled performance

Related Products
Agarose II (Low Melt)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Gel Electrophoresis
Melting PointStandard Melting Point
Product TypeAgarose I (Molecular Biology Grade)
Quantity100 g
Separation Range100 bp to >30 kb
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
Do you offer DMEM/F12 medium without folic acid?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
79423698Certificate of AnalysisApr 22, 202517850
79423699Certificate of AnalysisApr 22, 202517852
AA4587741Certificate of AnalysisFeb 12, 202517850, DNU-17850
AA4538121ACertificate of AnalysisFeb 12, 202517852, DNU-17852
AA4538121Certificate of AnalysisFeb 12, 202517852, DNU-17852
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection