Suero porcino
Suero porcino
Gibco™

Suero porcino

Estándar (esterilizado mediante filtro)
Have Questions?
Número de catálogoCantidad
26250084500 mL
Número de catálogo 26250084
Precio (EUR)
199,00
Each
-
Añadir al carro de la compra
Cantidad:
500 mL
Recurring order eligible. Learn more »
Precio (EUR)
199,00
Each
Añadir al carro de la compra
Ask our AI about this Product
  • Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • Sera Category: Specialty
  • Origin: New Zealand
  • Age: 1 year or less
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Especificaciones
Línea de productosGibco™
Condiciones de envíoCongelado
EspecieCerdo/porcino, porcino
AgeNormalmente menos de un año
País de origenNueva Zelanda
FormularioLíquido
Tipo de productoSuero porcino
Cantidad500 mL
Serum TreatmentEstándar (esterilizado mediante filtro)
EsterilidadEstéril
Unit SizeEach
Contenido y almacenamiento
Condiciones de almacenamiento: ≤10 °C
Condiciones de envío: Congelado
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
3220211Certificate of Analysis27 jun 202526250084
3128456Certificate of Analysis14 mar 202526250084
2540342Certificate of Analysis29 ene 202526250084
2828448Certificate of Analysis28 ene 202526250084
2888012Certificate of Analysis26 ene 202526250084
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Preguntas frecuentes

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

Other products to consider



Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection