Pierce™ ECL Plus Western Blotting Substrate
Pierce™ ECL Plus Western Blotting Substrate
Thermo Scientific™

Pierce™ ECL Plus Western Blotting Substrate

Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-rayRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensionsLxWxH
32132X3
32134
32132
Catalog number 32132X3
Price (USD)
817.65
Online Exclusive
922.00
Save 104.35 (11%)
Each
-
Add to cart
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
817.65
Online Exclusive
922.00
Save 104.35 (11%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196X3) through Fisher Scientific and other channels.

Features of ECL Plus Substrate:

Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
Higher sensitivity—detect targets down to the low-picogram level
Longer signal duration—sustained light output for as long as 5 hours
More imaging options—X-ray, CCD or laser-based imagers
More affordable—high quality and performance at a lower price than other suppliers' similar substrates

Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for western blot analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
SensitivityLow picogram to mid-femtogram
Sufficient For3000 cm2 of membrane
Detection MethodChemiluminescence, Fluorescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce™
Product TypeSubstrate
Quantity300 mL Kit
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product shipped on ice.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
AA403713ACertificate of AnalysisMay 13, 202532132
AD406099Certificate of AnalysisApr 23, 202532132
AB405125Certificate of AnalysisMar 07, 202532132
ZL403714ACertificate of AnalysisJan 31, 202532134
ZL403714Certificate of AnalysisJan 31, 202532134
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection