Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Thermo Scientific Chemicals

Hematoxylin, 85%, pure, certified

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 411160250
Price (USD)
98.65
Special offer
Online exclusive
Ends: 26-Sep-2025
122.00
Save 23.35 (19%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
98.65
Special offer
Online exclusive
Ends: 26-Sep-2025
122.00
Save 23.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS517-28-2
IUPAC Name(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Molecular FormulaC16H14O6
InChI KeyWZUVPPKBWHMQCE-WKTCHCBJNA-N
SMILES[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to brown
Loss on drying=<10 % (1 g, 105°C, 3 h)
Appearance (Form)Powder
UV spectrophotometry>=80 % (On dry basis)
BSC CertificationPasses
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472183Certificate of AnalysisMay 17, 2025411165000, 411161000, 411160250
A0464432Certificate of AnalysisJul 09, 2024411165000, 411161000, 411160250
A0459985Certificate of AnalysisFeb 12, 2024411165000, 411161000, 411160250
A0456721Certificate of AnalysisOct 04, 2023411165000, 411161000, 411160250
A0398481Certificate of AnalysisDec 14, 2022411165000, 411161000, 411160250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection