Search Thermo Fisher Scientific
Catalog Number | No. of Tests |
---|---|
A10628 | 25 |
A10346 | 100 |
When stored as directed (2-8 degrees C), this kit is stable for at least 1 year. Please do not freeze the kit. Please visit the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/arc_amine_reactive_comp_bead_kit_man.pdf) for further technical details.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
This can occur if the solution of beads was ever frozen. The formation of ice excludes the beads and compresses the beads together. Once frozen, the beads cannot be resuspended into solution and are no longer usable.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
This is because succinimidyl ester (NHS ester) groups are very labile to water; they dissociate readily from the dye structure if held in solvents that are contaminated with water, as is possible from condensation or absorbing water from the atmosphere.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Yes. The succinimidyl ester (NHS ester) and isothiocyanate (e.g., FITC, TRITC) dye derivatives may be attached to the ArC beads, but you would have to optimize the amount of reactive dye per ArC beads.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
It is possible. But if you have a lot of values over 100, you probably need to look at the voltages that you are using for your data collection.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Share catalog number, name or link