CellTracker™ Fluorescent Probes
CellTracker™ Fluorescent Probes
Invitrogen™

CellTracker™ Fluorescent Probes

CellTracker™ is a fluorescent dye well suited for monitoring cell movement or location. After loading into cells, the dye is well retained, allowing for multigenerational tracking of cellular movements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityDye Type
C3455120 x 50 μgCellTracker Orange CMRA
C21105 mgCellTracker™ Blue CMAC
C21115 mgCellTracker™ Blue CMHC
C128815 mgCellTracker Blue CMF2HC
C3456520 x 15 μgCellTracker™ Deep Red
C702520 x 50 μgCellTracker Green CMFDA
C29251 mgCellTracker Green CMFDA
C21025 mgCellTracker Green BODIPY
C29271 mgCellTracker Orange CMTMR
C3455220 x 50 μgCellTracker™ Red CMTPX
C100945 x 0.1 mgCellTracker Violet BMQC
Catalog number C34551
Price (USD)
543.65
Online Exclusive
560.00
Save 16.35 (3%)
Each
In stock
Add to cart
Quantity:
20 x 50 μg
Dye Type:
CellTracker Orange CMRA
Recurring order eligible. Learn more »
Price (USD)
543.65
Online Exclusive
560.00
Save 16.35 (3%)
Each
Add to cart
Ask our AI about this Product
Cell movement and location studies require specialized probes that are nontoxic to living cells and well retained, allowing for multigenerational tracking. The CellTracker fluorescent probes are available in a range of fluorescent colors to match instrument lasers and filters, and to accommodate co-staining with antibodies or other cell analysis probes. These dyes are excellent tools for monitoring cell movement, location, proliferation, migration, chemotaxis, and invasion.

Features of the CellTracker dyes include:

  • Easy to use—remove culture media, add dye, incubate 15-45 minutes, and image cells
  • Excellent retention—fluorescent signal retention of >72 hours (typically three to six generations)
  • Ideal tracking dyes—monitor cell movement, location, proliferation, migration, chemotaxis, and invasion
  • Low cytotoxicity—does not affect viability or proliferation
  • Cell Versatility– works with cells in suspension and adherent cells

The CellTracker fluorescent probes have been designed to freely pass through cell membranes; however, once inside the cell are transformed into cell-impermeant reaction products. The CellTracker fluorescence probes (except for CellTracker Deep Red) contain a chloromethyl or bromomethyl group that reacts with thiol groups, utilizing a glutathione S-transferase–mediated reaction. In most cells, glutathione levels are high (up to 10 mM) and glutathione transferase is ubiquitous. CellTracker Deep Red probe contains a succinimidyl ester reactive group, which reacts with amine groups present on proteins.

After conversion to impermeant versions, the CellTracker fluorescent probes are well retained in living cells through several generations. The probes are transferred to daughter cells, but are not transferred to adjacent cells in a population. Cells loaded with the CellTracker fluorescent probes display fluorescence for at least 72 hours and exhibit ideal tracking dye properties—they are stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH. Additionally, several CellTracker fluorescent probes with various excitation and emission spectra are available allowing for multiplexing in fluorescence imaging. The CellTracker dyes are retained with fixation and permeabilization, enabling their use with antibodies for fluorescence immunostaining applications.

Spectral characteristics of the fluorescent CellTracker probes:

  • CellTracker Blue CMAC—353/466 nm
  • CellTracker Blue CMF2HC—371/464 nm
  • CellTracker Blue CMHC—372/470 nm
  • CellTracker Violet BMQC—415/516 nm
  • CellTracker Green CMFDA—492/517 nm
  • CellTracker Green BODIPY™—522/529 nm
  • CellTracker Orange CMTMR—541/565 nm
  • CellTracker Orange CMR—548/576 nm
  • CellTracker Red CMTPX—577/602 nm
  • CellTracker Deep Red—630/660 nm

Fluorescent CellTracker reagents include the blue-fluorescent chloromethyl derivatives of amino-, hydroxy-, and difluorohydroxycoumarin (CMAC, CMHC and CMF2HC), the green-fluorescent chloromethyl derivatives of fluorescein diacetate (CMFDA) and a BODIPY™ dye, the orange-fluorescent CMTMR and CMRA, and the red-fluorescent CMTPX. CellTracker Blue CMAC, CMHC, and CMF2HC, CellTracker Violet, the violet-fluorescent bromomethyl derivative of coumarin (BMQC), CellTracker Green BODIPY™, CellTracker Orange CMTMR, and CellTracker Red CMTPX do not require enzymatic cleavage to activate their fluorescence, whereas the green CMFDA and orange CMRA do require enzymatic cleavage. The impermeable reaction products of the chloromethyl or bromomethyl coumarins have excellent retention, strong fluorescence, and relatively uniform cytoplasmic staining, making these derivatives potentially useful for correcting motion artifacts in imaging. CMFDA is colorless and non-fluorescent until cytosolic esterases cleave off the acetates, releasing a brightly fluorescent product.

In addition to bright fluorescence and excellent retention, the CellTracker fluorescent probes do not contribute to cytotoxicity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
DescriptionCellTracker™ Orange CMRA Dye, 20 x 50 μg
Dye TypeCellTracker Orange CMRA
Emission576 nm
Excitation Wavelength Range548 nm
FormDry Powder
Product LineCellTracker™
Quantity20 x 50 μg
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Shipping ConditionRoom Temperature
Label TypeOther Labels or Dyes
Product TypeDye
SubCellular LocalizationCytoplasm
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.
PG2624-PJT8477-COL022341-Media-Promo-Pod-Graphic-Global-F-278x123
four-plex-image-COS7-live-cell zoomed
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

I want to do a cell migration study for around 4 hours and need to fluorescently label the cells with a dye. What do you recommend?

Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I labeled my cells with Calcein, AM, but when I imaged the next day, there was no fluorescence from Calcein. Why?

Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.

For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can the CellTracker dyes be fixed?

Yes, the CellTracker dyes react with any accessible thiol part of the protein and can be fixed. However, some CellTracker dyes may be attached to small metabolites that can leak from the cell following permeabilization. This can result in decreased fluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

I stained two populations of cells, one with CellTracker Green and the other with CellTracker Red, but it looks like there may be crossover of the red dye to the green cells. What is going on?

One possibility is that there is spectral bleedthrough between the dyes. Be sure to check the single-color samples by imaging the red cells in green and imaging the green cells in red, using the optimal imaging settings for the other color. If you see bleedthrough with these controls, then you will have to reduce the dye label concentration to reduce the brightness of the dyes, or choose dyes that are farther apart spectrally. If the issue isn’t bleedthrough, another possibility is that the cells were not adequately washed after staining, allowing some unincorporated dye to remain and label the other cells after they were introduced. Extending washes and wash times should help with this.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I'm trying to stain my cells with CellTracker dyes or CFDA SE, but I'm not seeing much signal. What can I do?

First, make sure you aren’t staining in the presence of serum, since serum can have esterase activity that can prematurely cleave the AM group on these dyes, preventing entry into cells. After staining, it’s okay to return the cells to medium containing serum. After this, you can try increasing the concentration and label time to get a higher intensity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Figures

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3148384Certificate of AnalysisJun 18, 2025C34565
3154213Certificate of AnalysisMay 13, 2025C2925
3148319Certificate of AnalysisMay 04, 2025C7025
3123542Certificate of AnalysisApr 17, 2025C2927
3135768Certificate of AnalysisApr 08, 2025C12881
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (457)

Citations & References
Abstract
In situ image analysis of interactions between normal human keratinocytes and fibroblasts cultured in three-dimensional fibrin gels.
Authors:Sun T,Haycock J,Macneil S
Journal:Biomaterials
PubMed ID:16510181
Evaluation of a flow cytometric fluorescence quenching assay of phagocytosis of sensitized sheep erythrocytes by polymorphonuclear leukocytes.
Authors:Van Amersfoort ES, Van Strijp JA
Journal:Cytometry
PubMed ID:7875036
A number of reports have been published describing phagocytosis assays for flow cytometric analysis. In some of these, the fluorescence quenching technique has been used to discriminate between adherent and ingested particles. In this report, we have evaluated the efficacy of a quantitative fluorescence quenching technique with crystal violet and ... More
A point mutation in the binding subunit of a retroviral envelope protein arrests virus entry at hemifusion.
Authors:Zavorotinskaya T, Qian Z, Franks J, Albritton LM
Journal:J Virol
PubMed ID:14671127
The transmembrane subunits of viral envelope proteins are thought to perform all of the functions required for membrane fusion during entry of enveloped viruses. However, changes in a conserved SPHQ motif near the N terminus of the receptor binding subunit of a murine leukemia virus (MLV) envelope protein block infection ... More
Direct priming of antiviral CD8+ T cells in the peripheral interfollicular region of lymph nodes.
Authors:Hickman HD, Takeda K, Skon CN, Murray FR, Hensley SE, Loomis J, Barber GN, Bennink JR, Yewdell JW,
Journal:Nat Immunol
PubMed ID:18193049
'It is uncertain how antiviral lymphocytes are activated in draining lymph nodes, the site where adaptive immune responses are initiated. Here, using intravital microscopy we show that after infection of mice with vaccinia virus (a large DNA virus) or vesicular stomatitis virus (a small RNA virus), virions drained to the ... More
Single-molecule analysis of cadherin-mediated cell-cell adhesion.
Authors:Panorchan P, Thompson MS, Davis KJ, Tseng Y, Konstantopoulos K, Wirtz D
Journal:J Cell Sci
PubMed ID:16371651
'Cadherins are ubiquitous cell surface molecules that are expressed in virtually all solid tissues and localize at sites of cell-cell contact. Cadherins form a large and diverse family of adhesion molecules, which play a crucial role in a multitude of cellular processes, including cell-cell adhesion, motility, and cell sorting in ... More
457 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection