1x1 image pixel for data collection
T7 RNA Polymerase (20 U/μL)
T7 RNA Polymerase (20 U/μL)
Thermo Scientific™

T7 RNA Polymerase (20 U/μL)

Thermo Scientific Bacteriophage T7 RNA polymerase is a DNA-dependent RNA polymerase with strict specificity for its respective double-stranded promoters. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentrationQuantity
EP011120U/µL5,000 units
EP011220U/µL5 x 5,000 units
Catalog number EP0111
Price (USD)
109.00
Each
In stock
Add to cart
Concentration:
20U/µL
Quantity:
5,000 units
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
109.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Bacteriophage T7 RNA polymerase is a DNA-dependent RNA polymerase with strict specificity for its respective double-stranded promoters. It catalyzes the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.

Highlights

• Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Applications

Synthesis of unlabeled and labeled RNA that can be used:

• For hybridization, in vitro RNA translation
• As aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing
• In studies of RNA secondary structure and RNA-protein interactions, RNA splicing

Consensus promoter sequence:
T7: TAATACGACTCACTATAGGGAGA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20U/µL
Quantity5,000 units
PolymeraseT7 RNA Polymerase
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What is the advantage of using TranscriptAid T7 High Yield Transcription Kit over Thermo Scientific T7 RNA Polymerase with a standard transcription buffer?

TranscriptAid T7 High Yield Transcription Kit is designed to produce both long and short transcripts for applications that require large yields of RNA (milligram amounts). The kit can achieve 10 times greater amount than from conventional in vitro transcription reactions with T7 RNA polymerase due to its unique proprietary enzyme and buffer formulations. Please note that TranscriptAid T7 High Yield Transcription Kit is not recommended for generation of radioactively labelled RNA. Due to large quantities of RNA synthesized with the kit, generation of high specific activity radiolabeled probes would require prohibitively large amounts of radiolabeled nucleotide.

Does the T7 RNA Polymerase (20 U/μl) (Cat. No. EP0111) contain a His-Tag which could be used for purification or conjugation?

The T7 RNA Polymerase (20 U/μl) (Cat. No. EP0111) does not contain a His-Tag, and currently none of our T7 RNA polymerases have a tag available for purification/conjugation.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3273372Certificate of AnalysisJul 03, 2025EP0112
3245516Certificate of AnalysisMay 23, 2025EP0111
3245495Certificate of AnalysisMay 23, 2025EP0111
3226454Certificate of AnalysisApr 25, 2025EP0112
3161368Certificate of AnalysisFeb 03, 2025EP0111
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection