Pyrophosphatase, inorganic (0.1 U/μL)
Pyrophosphatase, inorganic (0.1 U/μL)
Thermo Scientific™

Pyrophosphatase, inorganic (0.1 U/μL)

Thermo Scientific Pyrophosphatase, Inorganic catalyzes the hydrolysis of inorganic pyrophosphate into two orthophosphates. The enzyme requires a divalent metal cationRead more
Have Questions?
Catalog NumberQuantity
EF022110 U
Catalog number EF0221
Price (USD)
70.65
Online Exclusive
89.75
Save 19.10 (21%)
Each
In stock
Add to cart
Quantity:
10 U
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
89.75
Save 19.10 (21%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pyrophosphatase, Inorganic catalyzes the hydrolysis of inorganic pyrophosphate into two orthophosphates. The enzyme requires a divalent metal cation with Mg2+ conferring the highest activity.

Highlights

• Active in Thermo Scientific buffers for DNA polymerases, RNA polymerases, and Reverse Transcriptases

Applications

• High yield RNA synthesis by in vitro transcription (see Reference 2)
• DNA polymerization reactions: preventing accumulation of pyrophosphate (see References 3, 4)
• Removal of contaminant PPi in reagents used for SNP genotyping by methods based on the detection of pyrophosphate

Note

The enzyme can be diluted with the supplied storage (dilution) buffer. Use of this enzyme in certain applications may be covered by patents and may require a license.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferStorage (Dilution) Buffer
Product TypePyrophosphatase
Quantity10 U
Concentration0.1 U/μL
EnzymePyrophosphatase
Unit SizeEach

Save 51% on various cell culture plasticware

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3276791Certificate of AnalysisJul 09, 2025EF0221
3249706Certificate of AnalysisMay 29, 2025EF0221
3195834Certificate of AnalysisMar 24, 2025EF0221
2817709Certificate of AnalysisJan 29, 2025EF0221
2790350Certificate of AnalysisJan 29, 2025EF0221
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection