Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200), Phenol Red Free
Product Image
Gibco™

Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200), Phenol Red Free

Human Large Vessel Endothelial Cell Basal Medium (formerly “Medium 200”) is a sterile, liquid tissue culture medium intended for useRead more
Have Questions?
Catalog NumberQuantity
M200PRF500500 mL
Catalog number M200PRF500
Price (USD)
107.00
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (USD)
107.00
Each
Add to cart
Ask our AI about this Product
Human Large Vessel Endothelial Cell Basal Medium (formerly “Medium 200”) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for normal human large vessel endothelial cells. When supplemented with low serum growth supplement (LSGS), this medium supports the plating and proliferation of endothelial cells at culture densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications may include primary isolation of endothelial cells. This medium is also available with phenol red (Cat. No. M200500).

Human Large Vessel Endothelial Cell Basal Medium is a basal culture medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of human large vessel endothelial cells, this medium must be supplemented with LSGS (Cat. No. S-003-10; kit = Cat. No. S-003-K).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEndothelial Cells
Product LineCascade Biologics™
Product TypeBasal Medium
Quantity500 mL
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHEPES
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
4 degrees C
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3118673Certificate of AnalysisMay 03, 2025M200PRF500
2556070Certificate of AnalysisJan 29, 2025M200PRF500
2562578Certificate of AnalysisJan 29, 2025M200PRF500
2785387Certificate of AnalysisJan 28, 2025M200PRF500
3025174Certificate of AnalysisJan 25, 2025M200PRF500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection