1x1 image pixel for data collection
Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200)
Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200)
Gibco™

Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200)

Human Large Vessel Endothelial Cell Basal Medium (formerly “Medium 200”) is a sterile, liquid tissue culture medium intended for useRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
M200500Promo Image500 mL
Catalog number M200500
Price (USD)
104.65
Online Exclusive
112.00
Save 7.35 (7%)
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (USD)
104.65
Online Exclusive
112.00
Save 7.35 (7%)
Each
Add to cart
Ask our AI about this Product
Human Large Vessel Endothelial Cell Basal Medium (formerly “Medium 200”) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for normal human large vessel endothelial cells. When supplemented with low serum growth supplement (LSGS), this medium supports the plating and proliferation of endothelial cells at culture densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications may include primary isolation of endothelial cells. This medium is also available without phenol red (Cat. No. M200PRF500).

Human Large Vessel Endothelial Cell Basal Medium is a basal culture medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of human large vessel endothelial cells, this medium must be supplemented with LSGS (Cat. No. S-003-10; kit = Cat. No. S-003-K).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEndothelial Cells
Product LineCascade Biologics™
Product TypeBasal Medium
Quantity500 mL
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHEPES, Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Media Formulations
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3102107Certificate of AnalysisFeb 27, 2025M200500
2375470Certificate of AnalysisJan 29, 2025M200500
2703618Certificate of AnalysisJan 29, 2025M200500
2753942Certificate of AnalysisJan 29, 2025M200500
2871872Certificate of AnalysisJan 27, 2025M200500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (1)

Citations & References
Abstract
Angiogenic properties of dehydrated human amnion/chorion allografts: therapeutic potential for soft tissue repair and regeneration.
Authors:Koob TJ, Lim JJ, Massee M, Zabek N, Rennert R, Gurtner G, Li WW,
Journal:
PubMed ID:24817999
Chronic wounds are associated with a number of deficiencies in critical wound healing processes, including growth factor signaling and neovascularization. Human-derived placental tissues are rich in regenerative cytokines and have been shown in randomized clinical trials to be effective for healing chronic wounds. In this study, PURION® Processed (MiMedx Group, ... More
1 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection