Human Melanocyte Growth Supplement-2 (HMGS-2), PMA-free
Product Image
Gibco™

Human Melanocyte Growth Supplement-2 (HMGS-2), PMA-free

Human Melanocyte Growth Supplement-2 (HMGS-2), PMA-free, is a sterile, concentrated (100X) solution intended for use with Medium 254 or MediumRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
S0165Promo Image5 mL
Catalog number S0165
Price (USD)
127.65
Online Exclusive
136.00
Save 8.35 (6%)
Each
Estimated availability date 07-Jul-2025
Add to cart
Quantity:
5 mL
Recurring order eligible. Learn more »
Price (USD)
127.65
Online Exclusive
136.00
Save 8.35 (6%)
Each
Add to cart
Ask our AI about this Product
Human Melanocyte Growth Supplement-2 (HMGS-2), PMA-free, is a sterile, concentrated (100X) solution intended for use with Medium 254 or Medium 254CF to culture human epidermal melanocytes. Each 5-mL bottle of HMGS-2 contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human epidermal melanocytes and is the correct amount of supplement for a 500-mL bottle of medium.

When a 500-mL bottle of medium is supplemented with HMGS-2, the final concentrations of the components in the supplemented medium are:

• Fetal bovine serum: 0.5%
• Basic fibroblast growth factor: 3 ng/mL
• Bovine pituitary extract: 0.2%
• Heparin: 3 μg/mL
• Hydrocortisone: 0.18 μg/mL
• Recombinant human insulin-like growth factor-I: 0.01 μg/mL
• Transferrin: 5 μg/mL
• Endothelin-1: 10 nM
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration100 X
Product LineCascade Biologics™
Quantity5 mL
Shipping ConditionDry Ice
SourceHuman
SpeciesHuman
Product TypeGrowth Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C).
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2397417Certificate of AnalysisJan 29, 2025S0165
2397437Certificate of AnalysisJan 29, 2025S0165
2787931Certificate of AnalysisJan 28, 2025S0165
2833383Certificate of AnalysisJan 27, 2025S0165
2930988Certificate of AnalysisJan 26, 2025S0165
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection