1x1 image pixel for data collection
Vybrant™ DiO Cell-Labeling Solution
Vybrant™ DiO Cell-Labeling Solution
Invitrogen™

Vybrant™ DiO Cell-Labeling Solution

The Vybrant™ DiO cell-labeling solution is a dye delivery solution that can be added directly to normal culture media toRead more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Catalog NumberQuantity
V22886Promo Image1 mL
Catalog number V22886
Price (USD)
196.65
Online Exclusive
202.00
Save 5.35 (3%)
1 mL
-
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Price (USD)
196.65
Online Exclusive
202.00
Save 5.35 (3%)
1 mL
Add to cart
Ask our AI about this Product
The Vybrant™ DiO cell-labeling solution is a dye delivery solution that can be added directly to normal culture media to uniformly label suspended or attached culture cells for use in cell-cell fusion, cellular adhesion and migration applications.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineVybrant™
Quantity1 mL
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeLiphophilic Tracer
SubCellular LocalizationCell Membranes, Lipids
Unit Size1 mL
Contents & Storage
Store at room temperature and protect from light.
Have questions about this product? Ask our AI assisted search.
I want to track my cells over time, and you have a lot of options to choose from. How do I pick the right one?
I want to perform a cell fusion assay, where one cell line is labeled with one color and the other cell line with another color, and combine with a nucleic acid stain. What do you recommend?
I need to look at live cell morphology deformation over the course of a few hours. What sort of membrane dye would be useful for this?
I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?
I want to track my cells with a nucleic acid stain, like DAPI or Hoechst dye. Do you recommend this?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3050026Certificate of AnalysisOct 31, 2024V22886
2836729Certificate of AnalysisFeb 16, 2024V22886
2652728Certificate of AnalysisJul 08, 2023V22886
2491432Certificate of AnalysisJun 30, 2022V22886
2373226Certificate of AnalysisAug 10, 2021V22886
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

A typical method is to label one cell line with orange fluorescent DiI C18 and the other cell line with green fluorescent DiO C18. These orange and green lipophilic cyanine dyes will stain the membranes of cells. Cells that fuse will then have both dyes, yielding a yellow color (when images are overlaid or cells are imaged in a dual-bandpass filter). These live cells can then be labeled with Hoechst 33342 (a cell-permeant blue DNA stain comparable in wavelength to DAPI), but only as an endpoint just before imaging (since DNA stains can interrupt DNA function).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Lipophilic cyanine dyes, such as DiI (Cat. No. D282), DiO (Cat. No. D275), DiD (Cat. No. D7757) or DiR (Cat. No. D12731), are commonly used. The longer the alkyl chain on the dye, the better the retention in lipophilic environments.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

This is not recommended. When these stains bind to DNA and RNA, they may affect the normal function of the nucleic acids, disrupting transcription, as well as replication. Other reagents, such as CellTracker dyes or Qtracker reagents are more optimized for tracking without disrupting normal activity. If a nuclear label is still desired, though, and the cells are mammalian and non-hematopoietic, CellLight nuclear reagents can transiently transfect cells to express GFP or RFP on a nuclear-expressing protein for up to several days without affecting function.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

Please see this Web link (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/cell-tracking.html) to help you choose the right option for your application. Start by planning how long you want to track your cells, then consider the mechanism of binding. Calcein dyes are very uniform in label and are good for short-term cell migration, but may be rapidly effluxed from some cell types. Lipophilic cyanine dyes, such as DiI, DiO, and similar dyes label cell membranes, don’t disrupt function, and can last longer, but have the potential to cross to other cells if membranes fuse. They are also lost upon permeabilization. CellTracker dyes are better for longer-term labeling, as they possess a mildly reactive chloromethyl moiety that allows covalent binding to cellular components. CFDA SE also covalently binds to cellular components. With all the reagents, their retention within cells is dependent upon the rate of cell division and the inherent properties of the cell (active efflux, membrane and protein turnover rates, etc.) and reagents that allow for covalent attachment exhibit longer retention than those that do not.

The longest-lasting and brightest options are the Qtracker reagents, which are taken up through endocytosis. These are so bright individual quantum dots can be detected, and are also robust enough to survive not only fixation and permeabilization, but even the heat and solvents used in paraffin processing.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (20)

Citations & References
Abstract
Submillisecond optical reporting of membrane potential in situ using a neuronal tracer dye.
Authors:Bradley J, Luo R, Otis TS, DiGregorio DA,
Journal:J Neurosci
PubMed ID:19625510
'A major goal in neuroscience is the development of optical reporters of membrane potential that are easy to use, have limited phototoxicity, and achieve the speed and sensitivity necessary for detection of individual action potentials in single neurons. Here we present a novel, two-component optical approach that attains these goals. ... More
Nanotubular highways for intercellular organelle transport.
Authors:Rustom A, Saffrich R, Markovic I, Walther P, Gerdes HH
Journal:Science
PubMed ID:14963329
Cell-to-cell communication is a crucial prerequisite for the development and maintenance of multicellular organisms. To date, diverse mechanisms of intercellular exchange of information have been documented, including chemical synapses, gap junctions, and plasmodesmata. Here, we describe highly sensitive nanotubular structures formed de novo between cells that create complex networks. These ... More
Sulfated glycosphingolipid as mediator of phagocytosis: SM4s enhances apoptotic cell clearance and modulates macrophage activity.
Authors:Popovic ZV, Sandhoff R, Sijmonsma TP, Kaden S, Jennemann R, Kiss E, Tone E, Autschbach F, Platt N, Malle E, Gröne HJ,
Journal:J Immunol
PubMed ID:17982067
Sulfoglycolipids are present on the surface of a variety of cells. The sulfatide SM4s is increased in lung, renal, and colon cancer and is associated with an adverse prognosis, possibly due to a low immunoreactivity of the tumor. As macrophages significantly contribute to the inflammatory infiltrate in malignancies, we postulated ... More
Surfactant protein-A and toll-like receptor-4 modulate immune functions of preterm baboon lung dendritic cell precursor cells.
Authors:Awasthi S, Madhusoodhanan R, Wolf R,
Journal:Cell Immunol
PubMed ID:21439559
Lung infections are important risk factors for an increased morbidity and mortality in prematurely-delivered babies. Immaturity of the innate immune components makes them extremely susceptible to infection. Recently, we isolated lung dendritic cell (DC)-precursor cells from preterm fetal baboons. The isolated cells were found to be defective in phagocytosing Escherichia ... More
Fluorescent dyes for lymphocyte migration and proliferation studies.
Authors:Parish CR
Journal:Immunol Cell Biol
PubMed ID:10571670
Fluorescent dyes are increasingly being exploited to track lymphocyte migration and proliferation. The present paper reviews the properties and performance of some 14 different fluorescent dyes that have been used during the last 20 years to monitor lymphocyte migration. Of the 14 dyes discussed, two stand out as being the ... More
20 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection