3D tumor spheroids on Nunclon Sphera 3D cell culture plates
Nunclon Sphera 3D culture system
Thermo Scientific’s Nunclon Sphera 3D culture system is designed for spheroid culture (e.g., 3D tumor spheroids) and organoid culture (e.g., brain organoids) with a wide selection of formats, from plates to dishes and flasks.
What are low attachment cell culture plates?
Low attachment cell culture plates, also known as spheroid culture plates, allow virtually no cell attachment to the cultureware, supporting the establishment of 3D spheroids and organoids via cell-to-cell aggregation through naturally secreted extracellular matrices (ECM).
Overview of the cell lines, culture conditions, and analysis techniques that are being used to transition from 2D to 3D cell models in the 3D Cell Culture & Analysis Handbook.
Have fun with mazes, lab awards, a word search, and more in the 3D Cell Culture & Analysis Activity Booklet.
Customer successes with Nunclon Sphera 3D cell culture system
Benefits of Nunclon Sphera ultra low attachment cell culture plates
Minimal cell adhesion in culture
Nunclon Sphera proprietary surface coating helps prevent protein adsorption to the cultureware surface, helping minimize monolayer cell adhesion to the culture vessel.
Bar chart showing reduced attachment for Nunclon Sphera surface
Figure 1. Compared to the standard cell culture flask (e.g., Nunclon Delta), minimal cell attachment was observed on the Nunclon Sphera 3D ultra low attachment culture flasks across three different cell types.
Consistent 3D tumor spheroid formation
The reliable cell growth and the ease in controlling spheroid size on the Nunclon Sphera 96-well U-bottom ultra low attachment 3D cell culture plate are important for applications where consistency and reproducibility are valued. 3D tumor spheroids imitate the in vivo development of solid tumors with homogenous growth and, in the cases of sizable tumors, the hypoxic core.
Microscopic views of early HCT 116 3D tumor spheroids cultured on Nunclon Sphera surface
Figure 2. Growth kinetics of HCT 116 colon cancer cells shows early spheroid formation in the Nunclon Sphera 3D cell culture plate, even with low initial seeding density of 100 cells/well.
Microscopic views of HCT 116 3D tumor spheroids at different seeding densities on Nunclon Sphera surface
Figure 3. Size of the HCT 116 3D tumor spheroids can be controlled by the initial seeding densities in the wells. HCT 116 colon cancer cells were cultured on the Nunclon Sphera 3D ultra low attachment cell culture plate for 112 hours.
Microscopic view of hypoxic core of HeLa 3D tumor spheroid cultured on Nunclon Sphera surface
Figure 4. The hypoxic core in a HeLa tumor spheroid after 2 days of culture on the Nunclon Sphera 96-well U-bottom spheroid culture plate with initial seeding density of 1,000 cells/well. The 3D tumor spheroid was stained with Invitrogen Image-iT Red Hypoxia Probe and Invitrogen NucBlue Live ReadyProbes reagent on a Nunc glass bottom dish.
Applications with Nunclon Sphera ultra low attachment cell culture plates
Neurosphere and brain organoid generation in Nunclon Sphera ultra low attachment plates
Neural stem cells (NSC) naturally secrete an extracellular matrix, allowing them to rapidly aggregate and form neurospheres. With the right 3D cell culture protocol, consistent cell growth of neurospheres leads to further differentiation to multi-cellular structures known as brain organoids with tissue-specific morphology and function.
Microscopic views of neurospheres growing on Nunclon Sphera surface
Figure 5. Human iPSC-derived neurospheres were cultured on a Nunclon Sphera spheroid culture plate for one week, stained with DAPI, and imaged.
Embryoid body formation using the Nunclon Sphera 3D culture system
Embryonic stem cells (ESC) and induced pluripotent stem cells (iPSC) form 3D aggregates, known as embryoid bodies, held together by a complex network of adhesion molecules. The paracrine signaling within an embryoid body is crucial to embryogenesis in response to environmental cues that lead to the eventual differentiation of three germ layers.
Microscopic view of viability of embryoid body cultured on Nunclon Sphera surface
Figure 6. Viability of human ESC in an embryoid body cultured on the Nunclon Sphera 3D cell culture dish in Gibco Essential 6 medium supplemented with TGF-b is evaluated by Invitrogen LIVE (green)/DEAD (red) viability assay (scale bar 1,000 μm).
Microscopic view of fluorescently stained germ layers in human embryoid bodies cultured on Nunclon Sphera surface
Figure 7. The differentiation of a human embryoid body to three germ layers on Nunc Lab Tek II Chamber Slides as shown by the expression of ectoderm marker beta-tubulin, endoderm marker alpha fetoprotein (AFP), and mesoderm marker smooth muscle actin (SMA). Counter-stained with DAPI.
Spheroid model system for high-throughput screening (HTS)
The high failure rate of new drug development suggests the need for better 3D cell models in the early stages of the drug development process. 3D cell spheroids and organoids of uniform size, centered in the wells of the Nunclon Sphera 3D cell culture microplates, present an attractive biologically relevant 3D cell model system for drug discovery and disease modeling amenable to high-throughput screening or high-content screening.
Microscopic view of A549 3D tumor spheroids in each well of 96-well Nunclon Sphera plate
Figure 8. A brightfield image of A549 human lung 3D tumor spheroids exhibiting uniform size and shape in the Nunclon Sphera 96-well U-bottom plate.
Microscopic view of fluorescently stained A549 and HeLa 3D tumor spheroids in 96-well Nunclon Sphera plate
Figure 9. Niclosamide-induced mitochondrial membrane depolarization and apoptosis are demonstrated using A549 and HeLa 3D tumor spheroids on the Nunclon Sphera 96-well U-bottom plate. Tumor spheroids are stained with MitoTracker Orange and CellEvent Green Caspase 3/7.
Ordering information for Nunclon Sphera ultra low attachment plates
Catalog # Name Size Price (USD) Qty
174925 96 Well Round (U) Bottom Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 8
206.65
Online Exclusive
224.00
Save 17.35 (8%)
174931 12 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
173.65
Online Exclusive
184.00
Save 10.35 (6%)
174930 24 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
164.65
Online Exclusive
177.00
Save 12.35 (7%)
174932 6 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
169.65
Online Exclusive
181.00
Save 11.35 (6%)
174927 96 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 8
219.65
Online Exclusive
234.00
Save 14.35 (6%)
174929 96 Well Round (U) Bottom Plate, Sphera Low-Attachment Surface, Pack of 5 Case of 50
1,090.65
Online Exclusive
1,192.00
Save 101.35 (9%)
Ordering information for Nunclon Sphera ultra low attachment flasks and dishes
Resources
Related products
Support
Additional resources
3D Cell Culture & Analysis Handbook
3D eLearning Module 
Learn about spheroid and organoid culture techniques, applications, and various imaging modalities including high-content analysis through this interactive online course
3D cell culture protocols
Data for cell lines on Nunclon surfaces
For Research Use Only. Not for use in diagnostic procedures.