quantstudio-5-real-time-pcr-system-human-identification-software-showcase-image-230x195
Enhanced data analysis for the QuantStudio 5 Real-Time PCR System for Human Identification
Applied Biosystems HID Real-Time PCR Analysis Software introduces the ability to analyze the sample data with a virtual standard curve defined by the user. In addition, the software includes the quantification, STR setup features, and STR data quality review functionality that integrate and streamline the quantification and STR analysis steps. By providing predefined templates for Quantifiler and STR kit runs, sample setup time is minimized. Incorporating HID-specific data-quality assessment and STR setup tools into a single software package enables faster, better decision making.
Order products
flexible-standard-curve-options-sml
 Click image to enlarge
Figure 1. Enhanced data analysis using virtual standard curve functionality
Flexible standard curve options
HID Real-Time PCR Analysis Software enables virtual standard curve functionality to minimize variation. This feature allows the user to define appropriate standard curve values (y-intercept and slope) for all targets applicable to the assay being run. The software allows for an enhanced data analysis pipeline that calculates the quantity of the samples using the traditional standards on the plate and/or virtual standard curve values (See Figure 1).
Streamlined HID-specific workflow
The software provides HID-specific templates preconfigured for the Quantifiler and DNA quantification kits, for a one-click start option. Customize and set up a run by selecting one of the Quantifiler assay template icons on the home screen (Figure 2).
In each template, predefined experimental properties, targets, sample types, and standard concentrations simplify the quantification run setup process. To set up a run, simply enter or import the sample names and well positions of any unknown samples.
streamlined-hid-specific-workflow-sm
 Click image to enlarge
Figure 2. The home screen provides direct access to HID-specific templates for all Quantifiler DNA Quantification Kits
The intuitive, easy-to-navigate user interface further streamlines the workflow  (Figure 3). The “Plate Setup”, “Run”, and “Analysis” functions are accessed in the “Experiment Menu” on the left navigation pane. Detailed experimental parameters and results are displayed conveniently on the right pane. For advanced data analysis, there are multiple ways to view and filter data, including plate map and table views, amplification plots, standard curve plots, multicomponent plots, and raw data views that support fast QC, sample data review, and troubleshooting.
For Research Use Only. Not for use in diagnostic procedures.