Automation
Microplate movers
The Orbitor RS2 Microplate Mover is a benchtop mover designed for use with the QuantStudio 12K Flex, QuantStudio 7 Pro, and QuantStudio 7 Flex real-time PCR systems, enabling real-time PCR in automated environments for increased sample throughput. 
Video Player is loading.
Current Time 0:00
Duration 1:38
Loaded: 0%
Stream Type LIVE
Remaining Time 1:38
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Evolves with your lab’s needs
    Allows a single or dual QuantStudio configuration with different plate storage options
    Advanced motion control
    Provides collision detection and position monitoring, allowing the mover to quickly resume without the need to re-home
    Momentum Workflow Scheduling Software
    Combines power and ease of use with an intuitive dashboard and logic for on-the-fly decisions, making it easy to use for novice or experienced users.
    Integrated barcode scanner
    Enables sample tracking and inventory management with a full 360⁰ workspace
    Available with two different plate storage options
    Choose the Orbitor RS2 Microplate Mover and applicable pre-configured storage bundle designed for the QuantStudio systems.
    Orbitor RS2 Microplate Mover
    Orbitor RS2 Microplate Mover Sequential Access Hotel Bundle
    Orbitor RS2 Microplate Mover Random Access Hotel Bundle
    Features
    Robust, efficient, and reliable benchtop mover with unmatched speed and accuracy
    Compatible with most SBS footprint plates
    Powered by Momentum Workflow Scheduling Software
    Holds up to 20 plates/3 stacks (96-well)
    Holds up to 40 plates/3 stacks (384-well)
    Sequential access hotels for plate storage and handling
    Provides the option to “run until empty”, which allows for additional plates to be added while the Orbitor RS2 is running
    Ideal for 0.1 mL, 0.2 mL 96-well plates and/or 384-well plates
    Random access hotels for plate storage and handling
    Holds up to 24 plates/3 hotels (384-well)
    Users can easily add new plates or change the order the plates are run
    Ideal for 384 well plates with multiple users
    Not ideal for 96-well plates
    Included
    Orbitor RS2 Microplate Mover
    Momentum Workflow Scheduling Software
    3 stacks
    Baseplate
    3 hotels
    Baseplate
    Catalog number
    A43780, A43779 (for the dual QuantStudio system integration) A43781
    A43782
    Required catalog numbers to create your automation bundle
    In addition to your QuantStudio 12K Flex, 7 Pro or 7 Flex System  
    Complete your Orbitor RS2 mover with the addition of either the Sequential Access Stack Bundle (Cat. No. A43781) or the Random Access Hotel Bundle (Cat.No. A43782).
    For integrating a dual QuantStudio system the integration kit (Cat. No. A43779) is required in addition to the automation bundle.
    A43780 + A43781 A43780 + A43782
    QuantStudio 12K Flex Accufill System
    The OpenArray Real-Time PCR Instrument and OpenArray AccuFill System can process hundreds to thousands of samples per day, enabling mid-density, high-throughput results with ease of use. The OpenArray AccuFill System accurately loads samples onto OpenArray plates, increasing reproducibility.
    The Accufill System comes included with the QuantStudio 12K Flex system to help you get the most out of your system.
    Upgrade kits
    Upgrade kits for the QuantStudio 6 and 7 Pro systems
    Increase application versatility with the following easy-to-change, powered-up interchangeable block types:
    QuantStudio Pro Block Upgrade Kit, 96-well, 0.1 mL
    QuantStudio Pro Block Upgrade Kit, 96-well, 0.2 mL
    QuantStudio Pro Block Upgrade Kit, 384-well
    QuantStudio 7 Pro Block Upgrade Kit, TaqMan Array Cards
    Features  
    Interchangeable 96-well block, 0.1 mL for the Pro systems
    Includes the 96-well 0.2-mL block, plus heated cover, and accessories
    Interchangeable 96-well block, 0.2 mL 
    Includes the 96-well 0.2-mL block, plus heated cover, and accessories
    Interchangeable 384-well block 
    Includes 384-well block, heated cover, and accessories
    Interchangeable block for use with TaqMan Array cards
    Includes the TAC block, plus heated cover and accessories
    Compatible systems
    QuantStudio 6 and 7 Pro systems
    QuantStudio 6 and 7 Pro systems
    QuantStudio 6 and 7 Pro systems
    QuantStudio 7 Pro System only
    Catalog number  
    A45955
    A45953
    A45954
    A45956
    Upgrade kits for the QuantStudio Flex systems
    QuantStudio 12K Flex OpenArray Block Upgrade Kit (with Accufill System) ViiA 7, QuantStudio 6 Flex, QuantStudio 7 Flex and QuantStudio 12k Flex System Block Upgrade Kit, 96 well
    ViiA 7, QuantStudio 6 Flex, QuantStudio 7 Flex and QuantStudio 12k Flex System Block Upgrade Kit, 384 well
    Features
    Accelerates genomic validation and screening, driving over 150,000 data points per workday
    Speeds up most projects to be completed in days instead of weeks
    Allows systems to operate using a 96-well block
    Includes standard 96-well block (0.2mL), heated cover, plate adaptor, and calibration kit
    Allows systems to operate using a 384-well block
    Includes 384-well block, heated cover, plate adaptor and calibration kit
    Compatible systems
    QuantStudio 12K Flex system
    QuantStudio 6 Flex, 7 Flex, and 12K Flex and ViiA 7 systems QuantStudio 6 Flex, 7 Flex, and 12K Flex and ViiA 7 systems
    Catalog number
    4471067
    4453543
    4453545
    Assays and reagents
    TaqMan Assays
    The perfect complement to any QuantStudio real-time PCR system.
    Applied Biosystems TaqMan Assays consist of target-specific primers and one or more probes optimized for specific applications including gene expression, SNP genotyping, miRNA, mutation detection, and copy number variation analysis. With more than 20 million predesigned assays, an easy-to-use design tool for custom assays, or custom primers and probes to develop your own…we have exactly what you need for successful qPCR. 
    Application-specific reagents optimized for QuantStudio instruments
    Applied Biosystems offers reagents featuring both TaqMan chemistry and SYBR Green chemistry optimized to work with all QuantStudio instruments.  Get the specificity and sensitivity you need for your applications.
    Want to evaluate our best performing master mixes?