wb-detection-reagents-showcase
With a variety of detection techniques for western blotting-chemiluminescence, fluorescence, or colorimetric, to choose from, you can select a technology to match your experimental requirements.
Chemiluminescent Detection Technical Tip
Fluorescent Detection Technical Tip
 Chemiluminescent detection
Chemiluminescent detection
Fluorescent detection
Fluorescent detection
Chromogenic detection
Chromogenic detection
Major advantageHighest sensitivityDetect multiple targets at onceEasily visualize blots without equipment
Technology overviewEnhanced chemiluminescent (ECL) HRP and AP substrates providing pictogram to femtogram level detectionSimultaneous detection of multiple proteins on the same blot using a variety of available fluorescent dyes and conjugated 2° antibodiesDirect visualization of your target protein using color detection reagents
Signal SourceIndirect signal from enzymatic reactionDirect signal from fluorophoreIndirect signal from enzymatic reaction
Signal DurationLimited (hours)Extended (weeks to months)Extended (weeks to months)
Sensitivity
Excellent, with a wide variety of substrates available
Good, may require higher concentration of secondary antibody
Limited, best for high abundant proteins
ConsistencyPossible variation between blots, can be mitigated by using high duration substratesHigh reproducibility between blotsPossible variation between blots
DetectionX-ray film and imaging instrumentsImaging instruments with appropriate filters or lasersVisual, no instrumentation required
QuantitationSingle-channel detection makes normalization challengingMultiplexing with an internal control makes normalization simplerSingle-channel detection makes normalization challenging
Other considerations
  • Stripping and reprobing of blot possible
  • Long exposure times possible, as no excitation light source required to capture signal
  • Care is needed to avoid fluorescence contamination
  • Longer exposure times can produce high background because of the small amount of excitation light passing through emission filters
  • Stripping and reprobing of blot not possible
  • Best application for high abundant proteins and when imaging or film processing instrumentation is not available
HRP substrates
Catalog # Name Size Price (USD) Qty
34577 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 200 mL Each
196.65
Online Exclusive
214.00
Save 17.35 (8%)
34578 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 1 L Each
561.65
Online Exclusive
624.00
Save 62.35 (10%)
34580 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 500 mL Each
338.65
Online Exclusive
378.00
Save 39.35 (10%)
34075 SuperSignal™ West Dura Extended Duration Substrate, 100 mL Each
395.65
Online Exclusive
428.00
Save 32.35 (8%)
34076 SuperSignal™ West Dura Extended Duration Substrate, 200 mL Each
703.65
Online Exclusive
756.00
Save 52.35 (7%)
34095 SuperSignal™ West Femto Maximum Sensitivity Substrate, 100 mL Each
428.65
Online Exclusive
475.00
Save 46.35 (10%)
34096 SuperSignal™ West Femto Maximum Sensitivity Substrate, 200 mL Each
701.65
Online Exclusive
774.00
Save 72.35 (9%)
32132 Pierce™ ECL Plus Western Blotting Substrate, 100 mL kit Each
377.65
Online Exclusive
426.00
Save 48.35 (11%)
32134 Pierce™ ECL Plus Western Blotting Substrate, 25 mL kit Each
125.65
Online Exclusive
142.00
Save 16.35 (12%)
32106 Pierce™ ECL Western Blotting Substrate Each
271.65
Online Exclusive
310.00
Save 38.35 (12%)
32109 Pierce™ ECL Western Blotting Substrate, 50 mL Each
78.65
Online Exclusive
89.50
Save 10.85 (12%)
32209 Pierce™ ECL Western Blotting Substrate, 250 mL Each
201.65
Online Exclusive
227.00
Save 25.35 (11%)
WP20004 Novex™ HRP Chromogenic Substrate (TMB) 250 mL
271.00
37574 1-Step™ Ultra TMB-Blotting Solution Each
237.65
Online Exclusive
266.00
Save 28.35 (11%)
34012 1-Step™ Chloronaphthol Substrate Solution Each
135.00
34065 Metal Enhanced DAB Substrate Kit Each
207.65
Online Exclusive
235.00
Save 27.35 (12%)
34000 Pierce™ CN/DAB Substrate Kit Each
184.00
A38554 SuperSignal™ West Atto Ultimate Sensitivity Substrate, 20 mL Each
139.65
Online Exclusive
150.00
Save 10.35 (7%)
A38555 SuperSignal™ West Atto Ultimate Sensitivity Substrate, 100 mL Each
444.65
Online Exclusive
482.00
Save 37.35 (8%)
AP substrates
Catalog # Name Size Price (USD) Qty
WP20002 Novex™ AP Chemiluminescent Substrate (CDP-Star™) 100 ml
631.65
Online Exclusive
686.00
Save 54.35 (8%)
WP20001 Novex™ AP Chromogenic Substrate (BCIP/ NBT) 250 mL
258.65
Online Exclusive
274.00
Save 15.35 (6%)
34042 1-Step™ NBT/BCIP Substrate Solution Each
218.00
34070 1-Step™ NBT/BCIP plus Suppressor Substrate Solution Each
161.00
Fluorescent secondary antibodies
Overview of western blot detection techniques
Chemiluminescent western blot detection
Chemiluminescent substrates are popular because they offer several advantages over other detection methods. These advantages have allowed chemiluminescence to become the detection method of choice in most protein laboratories. Using chemiluminescence allows multiple exposures to be made in order to obtain the best image. The detection reagents can be removed and the entire blot reprobed to visualize another protein or to optimize detection of the first protein. A large linear response range allows detection and quantitation for a large range of protein concentrations. Most importantly, chemiluminescence yields the greatest sensitivity of any available detection method.
Chemiluminescent Western Blot Detection
Key Highlights
  • Greatest sensitivity
  • Detection and quantitation for a large range of protein concentrations
Chemiluminescent substrates for horseradish peroxidase (HRP) are majority two-component systems consisting of a stable peroxide solution and an enhanced luminol solution. To make a working solution, the two components are mixed together. When incubated with a blot on which HRP-conjugated antibodies (or other probes) are bound, a chemical reaction emits light at 425 nm which can be captured with x-ray film, CCD camera imaging devices and phosphorimagers that detect chemiluminescence. Chemiluminescent substrates for alkaline phosphatase are also readily available. Click here for information on chemiluminescent western blot detection can be found here
Fluorescent western blot detection
Fluorescent reagents are growing in popularity for western blotting because they offer increased time savings over chemiluminescent detection and reduced chemical waste compared to both chemiluminescent or chromogenic detection systems. While the detection limits are still not as low as chemiluminescent detection, fluorescent detection has the unique advantage of allowing multiple targets to be assayed for on the same blot at the same time without the need to strip and reprobe.
Fluorescent Western Blot Detection
Key Highlights
  • Multiplexing capability
  • Direct signal
The growing demand for multiplex western blotting has driven the development of many new fluorescent dyes (such as Alexa Fluor Plus conjugates). These new fluorophores are brighter and more photo stable than the traditional fluorescein and rhodamine molecules traditionally used and comprise a broader range of non-overlapping spectra. Together with the advances in the digital imaging equipment these new fluorophores enable extremely powerful analyses in western blotting. Click here for information on fluorescent western blot detection can be found here.
Chromogenic western blot detection
Chromogenic or precipitating substrates have been used widely for many years and offer the simplest and most cost-effective method of western blot detection. When these substrates come in contact with the appropriate enzyme (e.g. Alkaline phosphatase- AP or horseradish peroxidase-HRP), they are converted to insoluble, colored products that precipitate onto the membrane. The resulting colored band or spot requires no special equipment for processing or visualizing. Chromogenic blotting substrates are available in a variety of specifications and formats, producing a range of colored precipitates. The appropriate substrate choice depends on the enzyme label and desired sensitivity. Similar to developing film, the blot is incubated in substrate until the desired amount of development is achieved. In contrast to chemiluminescent western blotting, the colored precipitate formed by chromogenic substrates cannot be easily stripped off to facilitate re-probing procedures. Therefore, it is important to allow the reaction to proceed until color development is satisfactory and then stop the reaction.
Chromogenic Western Blot Detection
Key Highlights
  • No special instrumentation required
  • Most cost-effective technique for high abundant targets
The low sensitivity of chromogenic substrates makes it difficult to optimize for detecting proteins of low abundance. Although the reaction can be allowed to develop for several hours or even overnight, this also allows background signal to continue to develop. Where chromogenic substrates fail in terms of sensitivity, they are ideal for applications where protein abundance is high. Click here for information on chromogenic western blot detection can be found here.  
 Chemiluminescent detection
Chemiluminescent detection
Fluorescent detection
Fluorescent detection
Chromogenic detection
Chromogenic detection
Major advantageHighest sensitivityDetect multiple targets at onceEasily visualize blots without equipment
Technology overviewEnhanced chemiluminescent (ECL) HRP and AP substrates providing pictogram to femtogram level detectionSimultaneous detection of multiple proteins on the same blot using a variety of available fluorescent dyes and conjugated 2° antibodiesDirect visualization of your target protein using color detection reagents
Signal SourceIndirect signal from enzymatic reactionDirect signal from fluorophoreIndirect signal from enzymatic reaction
Signal DurationLimited (hours)Extended (weeks to months)Extended (weeks to months)
Sensitivity
Excellent, with a wide variety of substrates available
Good, may require higher concentration of secondary antibody
Limited, best for high abundant proteins
ConsistencyPossible variation between blots, can be mitigated by using high duration substratesHigh reproducibility between blotsPossible variation between blots
DetectionX-ray film and imaging instrumentsImaging instruments with appropriate filters or lasersVisual, no instrumentation required
QuantitationSingle-channel detection makes normalization challengingMultiplexing with an internal control makes normalization simplerSingle-channel detection makes normalization challenging
Other considerations
  • Stripping and reprobing of blot possible
  • Long exposure times possible, as no excitation light source required to capture signal
  • Care is needed to avoid fluorescence contamination
  • Longer exposure times can produce high background because of the small amount of excitation light passing through emission filters
  • Stripping and reprobing of blot not possible
  • Best application for high abundant proteins and when imaging or film processing instrumentation is not available
HRP substrates
Catalog # Name Size Price (USD) Qty
34577 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 200 mL Each
196.65
Online Exclusive
214.00
Save 17.35 (8%)
34578 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 1 L Each
561.65
Online Exclusive
624.00
Save 62.35 (10%)
34580 SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 500 mL Each
338.65
Online Exclusive
378.00
Save 39.35 (10%)
34075 SuperSignal™ West Dura Extended Duration Substrate, 100 mL Each
395.65
Online Exclusive
428.00
Save 32.35 (8%)
34076 SuperSignal™ West Dura Extended Duration Substrate, 200 mL Each
703.65
Online Exclusive
756.00
Save 52.35 (7%)
34095 SuperSignal™ West Femto Maximum Sensitivity Substrate, 100 mL Each
428.65
Online Exclusive
475.00
Save 46.35 (10%)
34096 SuperSignal™ West Femto Maximum Sensitivity Substrate, 200 mL Each
701.65
Online Exclusive
774.00
Save 72.35 (9%)
32132 Pierce™ ECL Plus Western Blotting Substrate, 100 mL kit Each
377.65
Online Exclusive
426.00
Save 48.35 (11%)
32134 Pierce™ ECL Plus Western Blotting Substrate, 25 mL kit Each
125.65
Online Exclusive
142.00
Save 16.35 (12%)
32106 Pierce™ ECL Western Blotting Substrate Each
271.65
Online Exclusive
310.00
Save 38.35 (12%)
32109 Pierce™ ECL Western Blotting Substrate, 50 mL Each
78.65
Online Exclusive
89.50
Save 10.85 (12%)
32209 Pierce™ ECL Western Blotting Substrate, 250 mL Each
201.65
Online Exclusive
227.00
Save 25.35 (11%)
WP20004 Novex™ HRP Chromogenic Substrate (TMB) 250 mL
271.00
37574 1-Step™ Ultra TMB-Blotting Solution Each
237.65
Online Exclusive
266.00
Save 28.35 (11%)
34012 1-Step™ Chloronaphthol Substrate Solution Each
135.00
34065 Metal Enhanced DAB Substrate Kit Each
207.65
Online Exclusive
235.00
Save 27.35 (12%)
34000 Pierce™ CN/DAB Substrate Kit Each
184.00
A38554 SuperSignal™ West Atto Ultimate Sensitivity Substrate, 20 mL Each
139.65
Online Exclusive
150.00
Save 10.35 (7%)
A38555 SuperSignal™ West Atto Ultimate Sensitivity Substrate, 100 mL Each
444.65
Online Exclusive
482.00
Save 37.35 (8%)
AP substrates
Catalog # Name Size Price (USD) Qty
WP20002 Novex™ AP Chemiluminescent Substrate (CDP-Star™) 100 ml
631.65
Online Exclusive
686.00
Save 54.35 (8%)
WP20001 Novex™ AP Chromogenic Substrate (BCIP/ NBT) 250 mL
258.65
Online Exclusive
274.00
Save 15.35 (6%)
34042 1-Step™ NBT/BCIP Substrate Solution Each
218.00
34070 1-Step™ NBT/BCIP plus Suppressor Substrate Solution Each
161.00
Fluorescent secondary antibodies
Overview of western blot detection techniques
Chemiluminescent western blot detection
Chemiluminescent substrates are popular because they offer several advantages over other detection methods. These advantages have allowed chemiluminescence to become the detection method of choice in most protein laboratories. Using chemiluminescence allows multiple exposures to be made in order to obtain the best image. The detection reagents can be removed and the entire blot reprobed to visualize another protein or to optimize detection of the first protein. A large linear response range allows detection and quantitation for a large range of protein concentrations. Most importantly, chemiluminescence yields the greatest sensitivity of any available detection method.
Chemiluminescent Western Blot Detection
Key Highlights
  • Greatest sensitivity
  • Detection and quantitation for a large range of protein concentrations
Chemiluminescent substrates for horseradish peroxidase (HRP) are majority two-component systems consisting of a stable peroxide solution and an enhanced luminol solution. To make a working solution, the two components are mixed together. When incubated with a blot on which HRP-conjugated antibodies (or other probes) are bound, a chemical reaction emits light at 425 nm which can be captured with x-ray film, CCD camera imaging devices and phosphorimagers that detect chemiluminescence. Chemiluminescent substrates for alkaline phosphatase are also readily available. Click here for information on chemiluminescent western blot detection can be found here
Fluorescent western blot detection
Fluorescent reagents are growing in popularity for western blotting because they offer increased time savings over chemiluminescent detection and reduced chemical waste compared to both chemiluminescent or chromogenic detection systems. While the detection limits are still not as low as chemiluminescent detection, fluorescent detection has the unique advantage of allowing multiple targets to be assayed for on the same blot at the same time without the need to strip and reprobe.
Fluorescent Western Blot Detection
Key Highlights
  • Multiplexing capability
  • Direct signal
The growing demand for multiplex western blotting has driven the development of many new fluorescent dyes (such as Alexa Fluor Plus conjugates). These new fluorophores are brighter and more photo stable than the traditional fluorescein and rhodamine molecules traditionally used and comprise a broader range of non-overlapping spectra. Together with the advances in the digital imaging equipment these new fluorophores enable extremely powerful analyses in western blotting. Click here for information on fluorescent western blot detection can be found here.
Chromogenic western blot detection
Chromogenic or precipitating substrates have been used widely for many years and offer the simplest and most cost-effective method of western blot detection. When these substrates come in contact with the appropriate enzyme (e.g. Alkaline phosphatase- AP or horseradish peroxidase-HRP), they are converted to insoluble, colored products that precipitate onto the membrane. The resulting colored band or spot requires no special equipment for processing or visualizing. Chromogenic blotting substrates are available in a variety of specifications and formats, producing a range of colored precipitates. The appropriate substrate choice depends on the enzyme label and desired sensitivity. Similar to developing film, the blot is incubated in substrate until the desired amount of development is achieved. In contrast to chemiluminescent western blotting, the colored precipitate formed by chromogenic substrates cannot be easily stripped off to facilitate re-probing procedures. Therefore, it is important to allow the reaction to proceed until color development is satisfactory and then stop the reaction.
Chromogenic Western Blot Detection
Key Highlights
  • No special instrumentation required
  • Most cost-effective technique for high abundant targets
The low sensitivity of chromogenic substrates makes it difficult to optimize for detecting proteins of low abundance. Although the reaction can be allowed to develop for several hours or even overnight, this also allows background signal to continue to develop. Where chromogenic substrates fail in terms of sensitivity, they are ideal for applications where protein abundance is high. Click here for information on chromogenic western blot detection can be found here.  
For Research Use Only. Not for use in diagnostic procedures.