Cell lysis is a fundamental step in biological research that involves breaking open cell membranes to release cellular contents, allowing for the extraction and study of intracellular components such as proteins, DNA, RNA, and organelles. Cell lysis buffers are central to successful cell lysis. The careful selection of the most suitable lysis buffer is crucial for obtaining high-quality samples and accurate results in a wide range of applications. Cell lysis buffers are specially formulated to effectively disrupt cell membranes while maintaining the stability and activity of intracellular components. They typically contain detergents, salts, chelating agents, and protease inhibitors tailored to specific cell types and compatible with downstream applications such as western blot, ELISA, protein purification, immunoprecipitation, mass spectrometry, and gene expression analysis.
Thermo Fisher Scientific offers a range of lysis buffers specifically designed to address the distinct characteristics of mammalian, bacterial, plant, yeast, and insect samples. Mammalian cells commonly use RIPA buffer as it contains detergents and salts that can efficiently lyse cells and offer the flexibility to add protease inhibitors for maintaining the integrity of extracted proteins. Bacterial cells require B-PER or BugBuster which contains enzymes or detergents that effectively break down the bacterial cell wall. Specialized buffers are available for the unique characteristics of plant, yeast, and insect samples. Cell lysis buffers have been optimized to delicately disrupt cell membranes and extract cellular contents with precision and efficiency, while maintaining their integrity and functional activity. By understanding the types of lysis buffers available for different sample types, researchers can choose the lysis buffer that is most suitable for their experimental needs to help ensure reliable protein extraction and accurate downstream analysis. Choose from our comprehensive range of cell lysis buffers based on the specific needs of your experiment.
Key features of cell lysis buffers
  • Specificity—designed for specific sample types including mammalian cells, bacteria, plant, yeast, and insect cells helping ensure optimal performance
  • Efficiency—offer efficient and rapid cell membrane disruption helping ensure maximum recovery of intracellular content while minimizing sample loss
  • Preservation—preserve the integrity and functionality of proteins, DNA, RNA, and organelles preventing degradation and maintaining the functional activity
  • Compatibility—compatible with a variety of downstream applications, while safeguarding the quality of samples for downstream analysis
Choose the best lysis buffer for your experimental needs
Cell lysis buffers for mammalian cells often contain detergents, salts, and other solutions to disrupt the lipid bilayer of the cell membrane and extract proteins, DNA, RNA, and other cellular components. These lysis buffers include the option to add protease or phosphatase inhibitors to aid in stabilizing the extracted sample.
Cell lysis buffers for mammalian culture cells
 RIPA Lysis BufferIP Lysis BufferM-PER Mammalian Protein Extraction ReagentNP-40 Cell Lysis BufferCell Lysis Buffer
When to useExtraction of cytoplasmic and nuclear protein in denaturing conditionsFormulated specially for pull-down and immunoprecipitation
assays
Mild, non-denaturing and efficient lysis for cytoplasmic and nuclear protein extractionRecommended for extraction of cytoplasmic proteinsNeed a harsher buffer than NP-40 or when cytoplasmic and nuclear protein extraction is needed
Composition25 mM Tris-HCl, pH 7.6
150 mM NaCl
1% NP-40
1% sodium deoxycholate
0.1% SDS
A modified RIPA buffer formulation without SDSNon-denaturing detergent in 25 mM bicine buffer (pH 7.6)50 mM Tris, pH 7.4
250 mM NaCl
5 mM EDTA
50 mM NaF
1 mM Na3VO4
1% NP-40
0.02% NaN3
10 mM Tris, pH 7.4
100 mM NaCl
1 mM EDTA
1 mM EGTA
1 mM NaF
20 mM Na4P2O7
2 mM Na3VO4
1% Triton X-100
10% glycerol
0.1% SDS
0.5% deoxycholate
Recommended supplemental protease/phosphatase inhibitor(s)
Halt Protease Inhibitor Cocktail 
Halt Phosphatase Inhibitor Cocktail 
Halt Protease Inhibitor Cocktail 
Halt Phosphatase Inhibitor Cocktail 
Halt Protease Inhibitor Cocktail
Halt Protease Inhibitor Cocktail
PMSF Protease Inhibitor
Halt Protease Inhibitor Cocktail
PMSF Protease Inhibitor
Compatible protein assays
BCA assaysDetergent Compatible BradfordBCA assaysDetergent Compatible BradfordBCA assaysBradford assaysBCA assaysDetergent Compatible BradfordDetergent Compatible Bradford
Downstream compability
SDS-PAGE, western blottingImmunoprecipitations, pull-down assaysIP, protein electrophoresis, western blots, ELISA, EMSA, reporter assays, purification, activity assays, amine reactive labelingELISA, protein electrophoresis, western blottingELISA, protein electrophoresis, western blotting
Available size(s)
100 mL
250 mL
100 mL
250 mL
25 mL
250 mL
100 mL100 mL
User guide
User Guide: RIPA Lysis and Extraction BufferUser Guide: Pierce IP Lysis BufferUser Guide: M-PER Mammalian Protein Extraction ReagentUser Guide: NP-40 Cell Lysis BufferUser Guide: Cell Lysis Buffer
Cell lysis buffers for mammalian tissues
 T-PER Tissue Protein Extraction ReagentN-PER Neuronal Protein Extraction Reagent
When to use (compatible sample types)Heart, liver, kidney, lung, spleenBrain tissue and primary neurons
CompositionNon-denaturing detergent in 25 mM bicine, pH 7.6
150 mM NaCl
--
Compatible protein assaysBCA assays, Bradford assaysBCA assays, Detergent Compatible Bradford
Downstream compatibilityIP, western blots, ELISA, EMSA, purification, kinase assays, activity assays, amine reactive labelingIP, pulldowns, western blots, ELISA, enzyme assays, amine reactive labeling
Available sizes500 mL100 mL
User guideUser Guide: T-PER Tissue Protein Extraction ReagentUser Guide: N-PER Neuronal Protein Extraction Reagent
Mammalian cell lysis reagents for cultured cells
RIPA lysis buffer
RIPA (radioimmunoprecipitation assay) is a popular cell lysis option that allows for the extraction of membrane, cytoplasmic, and nuclear proteins from cells or tissues. This lysis buffer is particularly effective because of its non-ionic and ionic detergents such as SDS and sodium deoxycholate. Alternatively, these components may decrease activity of some protein kinases or enzymes, thus it is advised to prepare a RIPA buffer that does not contain SDS and sodium deoxycholate. Protease or phosphatase inhibitors can be utilized with RIPA buffer to prevent proteolysis and preserve the phosphorylation of proteins.
IP lysis buffer
Thermo Scientific Pierce IP lysis buffer is a whole cell lysis buffer based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. IP lysis buffer is an alternative cell lysis option to RIPA lysis buffer when a protein affinity purification experiment, such as immunoprecipitation (IP) or co-immunoprecipitation (CoIP), is the downstream application (Figure 1). This lysis buffer was formulated specially for pull-down and immunoprecipitation assays when it is especially important that the lysis reagent does not contain denaturants or components that might interfere with the antibody binding or protein interactions of interest. Protease or phosphatase inhibitors can be used to prevent proteolysis and maintain the phosphorylation of proteins.
IP and RIPA buffer lysates
Figure 1. Immunoprecipitation of cell lysates extracted using different lysis buffers. A549 cell lysates (200µg) were generated using different lysis buffers. EGFR and PP2A were immunoprecipitated using the Thermo Scientific Pierce Crosslink IP Kit and anti-EGFR antibody (Stressgen) or anti-PP2A antibody (Millipore). The immunoprecipitations were washed with the same lysis buffer used to lyse the cells. Lysates representing 5% of the total load (L) and elution (E) fractions were blotted with anti-EFGR or anti-PP2A antibody and using Thermo Scientific SuperSignal West Dura Substrate for detection.
M-PER mammalian protein extraction reagent
Thermo Scientific M-PER Mammalian Protein Extraction Reagent is an effective yet milder alternative to RIPA buffer. This reagent can extract soluble protein from primary, suspension, and adherent cell types (Figure 2). Lysis can be completed in only 5 minutes and directly added in culture plates, requiring little to no additional cellular disruption. M-PER can result in significantly more extracted protein compared with freeze/thaw and sonication methods (data not shown). Cell lysates obtained using M-PER are compatible with several downstream assays including immunoassays, enzyme assays, and various reporter assays. Additionally, M-PER contains dialyzable components without primary amines, making it suitable for luciferase, beta-galactosidase, and CAT assays unlike alternative protein extraction methods. 
Figure 2. Protein yield from various cell types using M-PER Mammalian Protein Extraction Reagent. Cells were harvested at 85% confluency, washed twice and collected in ice-cold PBS, and counted. For each cell type, 1 x 106 cells were pelleted by centrifugation at 2,000 x g for 5 minutes and lysed in 1 mL M-PER Reagent for 5 minutes. The cell lysates were clarified by centrifugation at 14,000 x g for 10 minutes, the supernatant was collected and the protein concentration (μg/million cells) was determined using the Pierce BCA Protein Assay.
Mammalian cell lysis reagents for tissues
Thermo Scientific T-PER Tissue Protein Extraction Reagent is designed for total protein extraction from mammalian tissue samples, including heart, liver, kidney, lung, and spleen (Figure 3). The T-PER reagent utilizes a non-denaturing detergent in 25 mM bicine, 150 mM NaCl (pH 7.6), and is used in conjunction with mechanical or manual homogenization. Furthermore, T-PER reagent was validated for protein extraction from fresh and frozen tissues.
Thermo Scientific N-PER Neuronal Protein Extraction Reagent is designed for total protein extraction from mammalian neuronal tissue samples or primary cultured neurons. N-PER lysis can be completed in less than 30 minutes and is used in conjunction with mechanical or manual homogenization. Protease or phosphatase inhibitors can be added to prevent proteolysis or protect against phosphatase activity normally present in brain tissue.
B-PER Bacterial Protein Extraction Reagents gently lyse E. coli and other species of bacterial cells and effectively extract soluble native and recombinant proteins. Several different ready-to-use formulations of B-PER reagent are available for different lysis needs. Depending on downstream application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added. 
 B-PER Complete Bacterial Protein Extraction ReagentB-PER Bacterial Protein Extraction ReagentB-PER II (2X) Bacterial Protein Extraction ReagentB-PER (PBS) Bacterial Protein Extraction Reagent
When to use (compatible sample types)E. coli, B. subtilisE. coli, B. subtilisE. coli, B. subtilisE. coli, B. subtilis
Features
  • Improved cell lysis and DNA digestion
  • Recovery of large molecular weight proteins
  • No enzyme components
  • Purification of affinity tagged proteins
  • Amine free formulation for direct compatibility of lysates with amine-reactive labeling and crosslinking
  • No enzyme components
  • Purification of proteins having low expression levels
  • No enzyme components
  • Amine free formulation for direct compatibility of lysates with amine-reactive labeling and crosslinking
For use with fresh and frozen cells?Yes, add EDTA to gram-negative fresh cellsYesYesYes
CompositionB-PER reagent formulated in Tris buffer with enzymes (lysozyme & universal nuclease)B-PER reagent formulated in Tris buffer2X B-PER reagent formulated in in Tris bufferB-PER reagent formulated in PBS
Nuclease and lysozyme in formulation?YesNoNoNo
Sample processing time15 min10–15 min10–15 min10–15 min
Mechanical disruption required?NoNo, but for gram-positive bacteria add enzymes for increased yieldNo, but for gram-positive bacteria add enzymes for increased yieldNo, but for gram-positive bacteria add enzymes for increased yield
Amount of sample processed100 g cell paste per 500 mL125 g cell paste per 500 mL125 g cell paste per 250 mL125 g cell paste per 500 mL
Compatible protein assaysBCA, Bradford assaysBCA, Bradford assaysBCA, Bradford assaysBCA, Bradford assays
Downstream compatibilitySDS-PAGE, western blotting, protein purificationSDS-PAGE, western blotting, protein purification (except for GST fusion proteins)SDS-PAGE, western blotting, protein purification (except for GST fusion proteins)SDS-PAGE, western blotting, protein purification (except for GST fusion proteins)
Available size(s)250 mL
500 mL
165 mL
250 mL
500 mL
250 mL500 mL
User guideUser Guide: B-PER Complete Bacterial Protein Extraction ReagentUser Guide: B-PER Bacterial Protein Extraction Reagent
B-PER Bacterial Protein Extraction Reagents
B-PER reagents can be used for various Gram-negative and Gram-positive bacteria such as S. aureus, H. pylori, and E. coli strains BL21(D3), JM109, DH5a, and M15. The reagent is also suitable for certain Archaebacteria species and cultured insect cells. Extraction does not require expensive equipment or mechanical disruption and can be performed in 10 to 15 minutes. B-PER reagent removes soluble protein from inclusion bodies and can be used to purify intact inclusion bodies whose less soluble proteins can be extracted by other means. 
B-PER complete bacterial protein extraction reagent
Thermo Scientific B-PER Complete Bacterial Protein Extraction Reagent is an all-in-one formulation that combines a lysis reagent with lysozyme and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria. Protein extracts are compatible with most protein assays and typical downstream applications such as affinity purification methods (e.g. GST, 6xHis) (Figure 5). 
bacterial protein purification comparison using B-PER and competitor
Figure 5. B-PER complete provides better compatibility with purification of 6His and GST fusion proteins. E. coli ER2566/pLATE51-Klenow, ER2566/pGST-CC-StpA and ER2566/pGST-Syk cell pellets cell pellets (0.5g), were resuspended in 2.5mL aliquots of B-PER Complete Reagent (B-PER) or competitor BugBuster master mix (Bug) with gentle vortexing for 15 minutes at room temperature. Insoluble cell debris was removed by centrifugation at 16,000 x g for 20 min at 4°C. 6xHis-Klenow protein was purified using Thermo Scientific HisPur Ni-NTA Agarose. GST-Syk and GST-StpA proteins were purified using Thermo Scientific Glutathione Agarose.
  • C = control (E. coli lysate from before induction with IPTG)
  • S = soluble protein (lysate after induction with 0.1mM IPTG)
  • E = elution fraction after protein purification
Specialized lysis buffers are available for plants, yeast, and insects that contain specific enzymes to break down their cell wall and extract the soluble protein. 
 Pierce Plant Total Protein Extraction KitY-PER Yeast Protein Extraction ReagentY-PER Plus Dialyzable Yeast Protein Extraction ReagentI-PER Insect Cell Protein Extraction Reagent
When to use (compatible sample types)Leaves, stems, soft roots, seedsSaccharomyces cerevisiae, Schizosaccharomyces pombe and Pichia pastorisSaccharomyces cerevisiae, Schizosaccharomyces pombe, Pichia pastoris, Bacillus subtilis, Escherichia coliSuspended or adherent cultured insect cells (Sf9, Sf21)
FeaturesObtain excellent protein yields in less than 10 minutes using a single buffer and a spin columnDoes not require glass beads for extractionFully dialyzable formulationGentle, effective protein extraction without sonication
Compatible protein assaysBCABCABCA, Bradford assaysBCA
Downstream compatibilitySDS or native-PAGE, western blotting, immunoprecipitation, affinity purification, activity assaysSDS-PAGE, western blotting, protein purification, activity assaysSDS-PAGE, western blotting, protein purificationWestern blotting, 6xHis-tagged protein purification, protein assays, ion-exchange chromatography
Available size(s)50 preps200 mL
500 mL
500 mL250 mL
User guideUser Guide: Pierce Plant Total Protein Extraction KitUser Guide: Y-PER Yeast Protein Extraction ReagentUser Guide: I-PER Insect Cell Protein Extraction Reagent
Plant tissue cell lysis kit
Plant cells are notoriously difficult to lyse and extract for proteomics work due to their tough cell walls and substantial polysaccharide content. The Thermo Scientific Pierce Plant Total Protein Extraction kit is composed of optimized buffers and filter cartridges that allow for efficient and rapid protein extraction in less than 10 minutes. The method has been validated for use with multiple plan organs (leaf, soft stem, root, seed, and flowers); multiple plant species (Arabidopsis, tobacco, maize, soybeans, peas, rice, and spinach); and fresh, frozen, and dehydrated tissue sources. The protein extracts can be used for applications such as SDS-PAGE, western blotting, immunoprecipitation, affinity purification, and activity assays.
Yeast cell lysis reagent
Thermo Scientific Y-PER Yeast Protein Extraction Reagent penetrates the tough yeast cell wall, perforating the cell wall and membrane and extracting soluble protein without completely damaging overall cell structure. Traditionally, yeast protein extraction required mechanical disruption with glass beads, making small-scale extraction difficult. However, Y-PER eliminated the need for glass bead lysis allowing for higher protein yield. Additionally, the elimination of glass bead methods allows for the protocol to be performed at room temperature and completed in 20 minutes. 
Two formulations are available depending on the downstream application. Y-PER reagent is high salt (>300 mM) and is effective for S. cerevisiae, S. pombe, C. albicans, and P. pastoris (as well as various gram-positive and gram-negative bacteria). Although the detergent is compatible with various downstream methods, it is not dialyzable, so it cannot be removed in cases where incompatibility exists. Thermo Scientific Y-PER Plus Dialyzable Yeast Protein Extraction Reagent is a phosphate-free, low-salt formulation with a dialyzable detergent, validated for use primarily with S. cerevisiae.
Insect cell lysis reagent
Thermo Scientific I-PER Insect Cell Protein Extraction Reagent enables gentle extraction of soluble protein from baculovirus-infected insect cells grown in suspension of monolayer culture (both Sf9 and Sf21 cells). The reagent maintains functionality of extracted proteins and is directly compatible with downstream applications, such as protein assays, western blotting, and His-tagged protein purification (IMAC).
Nucleases are commonly added to cell lysis reagents to remove the viscosity caused by the DNA content in bacterial cell lysates or to remove the DNA templates from RNAs produced by in vitro transcription. Lysozyme increases protein or nucleic acid extraction efficiency by breaking down the bacterial cell wall.
 Pierce Universal Nuclease for Cell LysisDNase I Solution (2500 U/mL)Micrococcal Nuclease Solution (≥100 U/µL)LysozymeLysozyme Solution
FeaturesDegrades all forms of DNA and RNADegrades both single-stranded and double-stranded DNADegrades nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experimentsBacterial cell wall lytic enzyme that improves protein or nucleic acid extraction efficiency
Available size(s)5 kU
100 kU
25 kU
0.5 mL150 µL5 g50 mg/mL
Cell lysis buffers for mammalian cells often contain detergents, salts, and other solutions to disrupt the lipid bilayer of the cell membrane and extract proteins, DNA, RNA, and other cellular components. These lysis buffers include the option to add protease or phosphatase inhibitors to aid in stabilizing the extracted sample.
Cell lysis buffers for mammalian culture cells
 RIPA Lysis BufferIP Lysis BufferM-PER Mammalian Protein Extraction ReagentNP-40 Cell Lysis BufferCell Lysis Buffer
When to useExtraction of cytoplasmic and nuclear protein in denaturing conditionsFormulated specially for pull-down and immunoprecipitation
assays
Mild, non-denaturing and efficient lysis for cytoplasmic and nuclear protein extractionRecommended for extraction of cytoplasmic proteinsNeed a harsher buffer than NP-40 or when cytoplasmic and nuclear protein extraction is needed
Composition25 mM Tris-HCl, pH 7.6
150 mM NaCl
1% NP-40
1% sodium deoxycholate
0.1% SDS
A modified RIPA buffer formulation without SDSNon-denaturing detergent in 25 mM bicine buffer (pH 7.6)50 mM Tris, pH 7.4
250 mM NaCl
5 mM EDTA
50 mM NaF
1 mM Na3VO4
1% NP-40
0.02% NaN3
10 mM Tris, pH 7.4
100 mM NaCl
1 mM EDTA
1 mM EGTA
1 mM NaF
20 mM Na4P2O7
2 mM Na3VO4
1% Triton X-100
10% glycerol
0.1% SDS
0.5% deoxycholate
Recommended supplemental protease/phosphatase inhibitor(s)
Halt Protease Inhibitor Cocktail 
Halt Phosphatase Inhibitor Cocktail 
Halt Protease Inhibitor Cocktail 
Halt Phosphatase Inhibitor Cocktail 
Halt Protease Inhibitor Cocktail
Halt Protease Inhibitor Cocktail
PMSF Protease Inhibitor
Halt Protease Inhibitor Cocktail
PMSF Protease Inhibitor
Compatible protein assays
BCA assaysDetergent Compatible BradfordBCA assaysDetergent Compatible BradfordBCA assaysBradford assaysBCA assaysDetergent Compatible BradfordDetergent Compatible Bradford
Downstream compability
SDS-PAGE, western blottingImmunoprecipitations, pull-down assaysIP, protein electrophoresis, western blots, ELISA, EMSA, reporter assays, purification, activity assays, amine reactive labelingELISA, protein electrophoresis, western blottingELISA, protein electrophoresis, western blotting
Available size(s)
100 mL
250 mL
100 mL
250 mL
25 mL
250 mL
100 mL100 mL
User guide
User Guide: RIPA Lysis and Extraction BufferUser Guide: Pierce IP Lysis BufferUser Guide: M-PER Mammalian Protein Extraction ReagentUser Guide: NP-40 Cell Lysis BufferUser Guide: Cell Lysis Buffer
Cell lysis buffers for mammalian tissues
 T-PER Tissue Protein Extraction ReagentN-PER Neuronal Protein Extraction Reagent
When to use (compatible sample types)Heart, liver, kidney, lung, spleenBrain tissue and primary neurons
CompositionNon-denaturing detergent in 25 mM bicine, pH 7.6
150 mM NaCl
--
Compatible protein assaysBCA assays, Bradford assaysBCA assays, Detergent Compatible Bradford
Downstream compatibilityIP, western blots, ELISA, EMSA, purification, kinase assays, activity assays, amine reactive labelingIP, pulldowns, western blots, ELISA, enzyme assays, amine reactive labeling
Available sizes500 mL100 mL
User guideUser Guide: T-PER Tissue Protein Extraction ReagentUser Guide: N-PER Neuronal Protein Extraction Reagent
Mammalian cell lysis reagents for cultured cells
RIPA lysis buffer
RIPA (radioimmunoprecipitation assay) is a popular cell lysis option that allows for the extraction of membrane, cytoplasmic, and nuclear proteins from cells or tissues. This lysis buffer is particularly effective because of its non-ionic and ionic detergents such as SDS and sodium deoxycholate. Alternatively, these components may decrease activity of some protein kinases or enzymes, thus it is advised to prepare a RIPA buffer that does not contain SDS and sodium deoxycholate. Protease or phosphatase inhibitors can be utilized with RIPA buffer to prevent proteolysis and preserve the phosphorylation of proteins.
IP lysis buffer
Thermo Scientific Pierce IP lysis buffer is a whole cell lysis buffer based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. IP lysis buffer is an alternative cell lysis option to RIPA lysis buffer when a protein affinity purification experiment, such as immunoprecipitation (IP) or co-immunoprecipitation (CoIP), is the downstream application (Figure 1). This lysis buffer was formulated specially for pull-down and immunoprecipitation assays when it is especially important that the lysis reagent does not contain denaturants or components that might interfere with the antibody binding or protein interactions of interest. Protease or phosphatase inhibitors can be used to prevent proteolysis and maintain the phosphorylation of proteins.
IP and RIPA buffer lysates
Figure 1. Immunoprecipitation of cell lysates extracted using different lysis buffers. A549 cell lysates (200µg) were generated using different lysis buffers. EGFR and PP2A were immunoprecipitated using the Thermo Scientific Pierce Crosslink IP Kit and anti-EGFR antibody (Stressgen) or anti-PP2A antibody (Millipore). The immunoprecipitations were washed with the same lysis buffer used to lyse the cells. Lysates representing 5% of the total load (L) and elution (E) fractions were blotted with anti-EFGR or anti-PP2A antibody and using Thermo Scientific SuperSignal West Dura Substrate for detection.
M-PER mammalian protein extraction reagent
Thermo Scientific M-PER Mammalian Protein Extraction Reagent is an effective yet milder alternative to RIPA buffer. This reagent can extract soluble protein from primary, suspension, and adherent cell types (Figure 2). Lysis can be completed in only 5 minutes and directly added in culture plates, requiring little to no additional cellular disruption. M-PER can result in significantly more extracted protein compared with freeze/thaw and sonication methods (data not shown). Cell lysates obtained using M-PER are compatible with several downstream assays including immunoassays, enzyme assays, and various reporter assays. Additionally, M-PER contains dialyzable components without primary amines, making it suitable for luciferase, beta-galactosidase, and CAT assays unlike alternative protein extraction methods. 
Figure 2. Protein yield from various cell types using M-PER Mammalian Protein Extraction Reagent. Cells were harvested at 85% confluency, washed twice and collected in ice-cold PBS, and counted. For each cell type, 1 x 106 cells were pelleted by centrifugation at 2,000 x g for 5 minutes and lysed in 1 mL M-PER Reagent for 5 minutes. The cell lysates were clarified by centrifugation at 14,000 x g for 10 minutes, the supernatant was collected and the protein concentration (μg/million cells) was determined using the Pierce BCA Protein Assay.
Mammalian cell lysis reagents for tissues
Thermo Scientific T-PER Tissue Protein Extraction Reagent is designed for total protein extraction from mammalian tissue samples, including heart, liver, kidney, lung, and spleen (Figure 3). The T-PER reagent utilizes a non-denaturing detergent in 25 mM bicine, 150 mM NaCl (pH 7.6), and is used in conjunction with mechanical or manual homogenization. Furthermore, T-PER reagent was validated for protein extraction from fresh and frozen tissues.
Thermo Scientific N-PER Neuronal Protein Extraction Reagent is designed for total protein extraction from mammalian neuronal tissue samples or primary cultured neurons. N-PER lysis can be completed in less than 30 minutes and is used in conjunction with mechanical or manual homogenization. Protease or phosphatase inhibitors can be added to prevent proteolysis or protect against phosphatase activity normally present in brain tissue.
B-PER Bacterial Protein Extraction Reagents gently lyse E. coli and other species of bacterial cells and effectively extract soluble native and recombinant proteins. Several different ready-to-use formulations of B-PER reagent are available for different lysis needs. Depending on downstream application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added. 
 B-PER Complete Bacterial Protein Extraction ReagentB-PER Bacterial Protein Extraction ReagentB-PER II (2X) Bacterial Protein Extraction ReagentB-PER (PBS) Bacterial Protein Extraction Reagent
When to use (compatible sample types)E. coli, B. subtilisE. coli, B. subtilisE. coli, B. subtilisE. coli, B. subtilis
Features
  • Improved cell lysis and DNA digestion
  • Recovery of large molecular weight proteins
  • No enzyme components
  • Purification of affinity tagged proteins
  • Amine free formulation for direct compatibility of lysates with amine-reactive labeling and crosslinking
  • No enzyme components
  • Purification of proteins having low expression levels
  • No enzyme components
  • Amine free formulation for direct compatibility of lysates with amine-reactive labeling and crosslinking
For use with fresh and frozen cells?Yes, add EDTA to gram-negative fresh cellsYesYesYes
CompositionB-PER reagent formulated in Tris buffer with enzymes (lysozyme & universal nuclease)B-PER reagent formulated in Tris buffer2X B-PER reagent formulated in in Tris bufferB-PER reagent formulated in PBS
Nuclease and lysozyme in formulation?YesNoNoNo
Sample processing time15 min10–15 min10–15 min10–15 min
Mechanical disruption required?NoNo, but for gram-positive bacteria add enzymes for increased yieldNo, but for gram-positive bacteria add enzymes for increased yieldNo, but for gram-positive bacteria add enzymes for increased yield
Amount of sample processed100 g cell paste per 500 mL125 g cell paste per 500 mL125 g cell paste per 250 mL125 g cell paste per 500 mL
Compatible protein assaysBCA, Bradford assaysBCA, Bradford assaysBCA, Bradford assaysBCA, Bradford assays
Downstream compatibilitySDS-PAGE, western blotting, protein purificationSDS-PAGE, western blotting, protein purification (except for GST fusion proteins)SDS-PAGE, western blotting, protein purification (except for GST fusion proteins)SDS-PAGE, western blotting, protein purification (except for GST fusion proteins)
Available size(s)250 mL
500 mL
165 mL
250 mL
500 mL
250 mL500 mL
User guideUser Guide: B-PER Complete Bacterial Protein Extraction ReagentUser Guide: B-PER Bacterial Protein Extraction Reagent
B-PER Bacterial Protein Extraction Reagents
B-PER reagents can be used for various Gram-negative and Gram-positive bacteria such as S. aureus, H. pylori, and E. coli strains BL21(D3), JM109, DH5a, and M15. The reagent is also suitable for certain Archaebacteria species and cultured insect cells. Extraction does not require expensive equipment or mechanical disruption and can be performed in 10 to 15 minutes. B-PER reagent removes soluble protein from inclusion bodies and can be used to purify intact inclusion bodies whose less soluble proteins can be extracted by other means. 
B-PER complete bacterial protein extraction reagent
Thermo Scientific B-PER Complete Bacterial Protein Extraction Reagent is an all-in-one formulation that combines a lysis reagent with lysozyme and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria. Protein extracts are compatible with most protein assays and typical downstream applications such as affinity purification methods (e.g. GST, 6xHis) (Figure 5). 
bacterial protein purification comparison using B-PER and competitor
Figure 5. B-PER complete provides better compatibility with purification of 6His and GST fusion proteins. E. coli ER2566/pLATE51-Klenow, ER2566/pGST-CC-StpA and ER2566/pGST-Syk cell pellets cell pellets (0.5g), were resuspended in 2.5mL aliquots of B-PER Complete Reagent (B-PER) or competitor BugBuster master mix (Bug) with gentle vortexing for 15 minutes at room temperature. Insoluble cell debris was removed by centrifugation at 16,000 x g for 20 min at 4°C. 6xHis-Klenow protein was purified using Thermo Scientific HisPur Ni-NTA Agarose. GST-Syk and GST-StpA proteins were purified using Thermo Scientific Glutathione Agarose.
  • C = control (E. coli lysate from before induction with IPTG)
  • S = soluble protein (lysate after induction with 0.1mM IPTG)
  • E = elution fraction after protein purification
Specialized lysis buffers are available for plants, yeast, and insects that contain specific enzymes to break down their cell wall and extract the soluble protein. 
 Pierce Plant Total Protein Extraction KitY-PER Yeast Protein Extraction ReagentY-PER Plus Dialyzable Yeast Protein Extraction ReagentI-PER Insect Cell Protein Extraction Reagent
When to use (compatible sample types)Leaves, stems, soft roots, seedsSaccharomyces cerevisiae, Schizosaccharomyces pombe and Pichia pastorisSaccharomyces cerevisiae, Schizosaccharomyces pombe, Pichia pastoris, Bacillus subtilis, Escherichia coliSuspended or adherent cultured insect cells (Sf9, Sf21)
FeaturesObtain excellent protein yields in less than 10 minutes using a single buffer and a spin columnDoes not require glass beads for extractionFully dialyzable formulationGentle, effective protein extraction without sonication
Compatible protein assaysBCABCABCA, Bradford assaysBCA
Downstream compatibilitySDS or native-PAGE, western blotting, immunoprecipitation, affinity purification, activity assaysSDS-PAGE, western blotting, protein purification, activity assaysSDS-PAGE, western blotting, protein purificationWestern blotting, 6xHis-tagged protein purification, protein assays, ion-exchange chromatography
Available size(s)50 preps200 mL
500 mL
500 mL250 mL
User guideUser Guide: Pierce Plant Total Protein Extraction KitUser Guide: Y-PER Yeast Protein Extraction ReagentUser Guide: I-PER Insect Cell Protein Extraction Reagent
Plant tissue cell lysis kit
Plant cells are notoriously difficult to lyse and extract for proteomics work due to their tough cell walls and substantial polysaccharide content. The Thermo Scientific Pierce Plant Total Protein Extraction kit is composed of optimized buffers and filter cartridges that allow for efficient and rapid protein extraction in less than 10 minutes. The method has been validated for use with multiple plan organs (leaf, soft stem, root, seed, and flowers); multiple plant species (Arabidopsis, tobacco, maize, soybeans, peas, rice, and spinach); and fresh, frozen, and dehydrated tissue sources. The protein extracts can be used for applications such as SDS-PAGE, western blotting, immunoprecipitation, affinity purification, and activity assays.
Yeast cell lysis reagent
Thermo Scientific Y-PER Yeast Protein Extraction Reagent penetrates the tough yeast cell wall, perforating the cell wall and membrane and extracting soluble protein without completely damaging overall cell structure. Traditionally, yeast protein extraction required mechanical disruption with glass beads, making small-scale extraction difficult. However, Y-PER eliminated the need for glass bead lysis allowing for higher protein yield. Additionally, the elimination of glass bead methods allows for the protocol to be performed at room temperature and completed in 20 minutes. 
Two formulations are available depending on the downstream application. Y-PER reagent is high salt (>300 mM) and is effective for S. cerevisiae, S. pombe, C. albicans, and P. pastoris (as well as various gram-positive and gram-negative bacteria). Although the detergent is compatible with various downstream methods, it is not dialyzable, so it cannot be removed in cases where incompatibility exists. Thermo Scientific Y-PER Plus Dialyzable Yeast Protein Extraction Reagent is a phosphate-free, low-salt formulation with a dialyzable detergent, validated for use primarily with S. cerevisiae.
Insect cell lysis reagent
Thermo Scientific I-PER Insect Cell Protein Extraction Reagent enables gentle extraction of soluble protein from baculovirus-infected insect cells grown in suspension of monolayer culture (both Sf9 and Sf21 cells). The reagent maintains functionality of extracted proteins and is directly compatible with downstream applications, such as protein assays, western blotting, and His-tagged protein purification (IMAC).
Nucleases are commonly added to cell lysis reagents to remove the viscosity caused by the DNA content in bacterial cell lysates or to remove the DNA templates from RNAs produced by in vitro transcription. Lysozyme increases protein or nucleic acid extraction efficiency by breaking down the bacterial cell wall.
 Pierce Universal Nuclease for Cell LysisDNase I Solution (2500 U/mL)Micrococcal Nuclease Solution (≥100 U/µL)LysozymeLysozyme Solution
FeaturesDegrades all forms of DNA and RNADegrades both single-stranded and double-stranded DNADegrades nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experimentsBacterial cell wall lytic enzyme that improves protein or nucleic acid extraction efficiency
Available size(s)5 kU
100 kU
25 kU
0.5 mL150 µL5 g50 mg/mL
Ordering information
Catalog # Name Size Price (USD) Qty
89900 RIPA Lysis and Extraction Buffer, 100 mL Each
115.65
Online exclusive
129.00
Save 13.35 (10%)
89901 RIPA Lysis and Extraction Buffer 250 mL
242.65
Online exclusive
272.00
Save 29.35 (11%)
87787 Pierce™ IP Lysis Buffer, 100 mL Each
109.65
Online exclusive
121.00
Save 11.35 (9%)
87788 Pierce™ IP Lysis Buffer, 250 mL Each
232.65
Online exclusive
257.00
Save 24.35 (9%)
78501 M-PER™ Mammalian Protein Extraction Reagent, 250 mL Each
351.65
Online exclusive
390.00
Save 38.35 (10%)
78503 M-PER™ Mammalian Protein Extraction Reagent, 25 mL Each
89.65
Online exclusive
104.00
Save 14.35 (14%)
FNN0021 Cell Lysis Buffer II Each
179.65
Online exclusive
187.00
Save 7.35 (4%)
FNN0011 Cell Extraction Buffer Each
177.65
Online exclusive
185.00
Save 7.35 (4%)
Catalog # Name Size Price (USD) Qty
89821 B-PER™ Complete Bacterial Protein Extraction Reagent, 250 mL Each
384.65
Online exclusive
427.00
Save 42.35 (10%)
89822 B-PER™ Complete Bacterial Protein Extraction Reagent, 500 mL Each
694.65
Online exclusive
770.00
Save 75.35 (10%)
78243 B-PER™ Bacterial Protein Extraction Reagent, 165 mL Each
213.65
Online exclusive
248.00
Save 34.35 (14%)
90084 B-PER™ Bacterial Protein Extraction Reagent, 250 mL Each
277.65
Online exclusive
310.00
Save 32.35 (10%)
78248 B-PER™ Bacterial Protein Extraction Reagent, 500 mL Each
378.65
Online exclusive
436.00
Save 57.35 (13%)
78260 B-PER™ II Bacterial Protein Extraction Reagent (2X) Each
414.65
Online exclusive
453.00
Save 38.35 (8%)
78266 B-PER™ Reagent (in Phosphate Buffer) Each
403.65
Online exclusive
453.00
Save 49.35 (11%)
Catalog # Name Size Price (USD) Qty
A44056 Pierce Plant Total Protein Extraction Kit Each
323.65
Online exclusive
337.00
Save 13.35 (4%)
78991 Y-PER™ Yeast Protein Extraction Reagent, 200 mL Each
215.65
Online exclusive
234.00
Save 18.35 (8%)
78990 Y-PER™ Yeast Protein Extraction Reagent, 500 mL Each
460.65
Online exclusive
500.00
Save 39.35 (8%)
78999 Y-PER™ Plus Dialyzable Yeast Protein Extraction Reagent Each
489.00
89802 I-PER™ Insect Cell Protein Extraction Reagent Each
308.65
Online exclusive
331.00
Save 22.35 (7%)
Catalog # Name Size Price (USD) Qty
88700 Pierce™ Universal Nuclease for Cell Lysis 5 kU
71.65
Online exclusive
78.50
Save 6.85 (9%)
88701 Pierce™ Universal Nuclease for Cell Lysis, 25 kU Each
220.65
Online exclusive
263.00
Save 42.35 (16%)
88702 Pierce™ Universal Nuclease for Cell Lysis, 100 kU Each
480.65
Online exclusive
560.00
Save 79.35 (14%)
90083 DNase I Solution (2500 U/mL) Each
120.65
Online exclusive
133.00
Save 12.35 (9%)
88216 Micrococcal Nuclease Solution (≥100 U/μL) Each
175.65
Online exclusive
191.00
Save 15.35 (8%)
90082 Lysozyme Solution (50 mg/mL) Each
78.50
Discover protein preparation tools
Protein sample preparation technical handbook
The Protein Preparation Handbook provides useful information on our broad selection of reagents and tools for protein extraction, clean-up, immunoprecipitation, and purification. Practical information, selection guides, and relevant data are included to help you improve your protein yield and downstream analysis.
protein preparation handbook
Protein sample preparation eLearning course
This modular, animated, and narrated eLearning course was developed to provide a succinct, contextual summary of common laboratory methods and techniques required to achieve optimal results in protein assays and experiments. There are knowledge checks throughout the course to test what you have learned.
eLearning courses
Stylesheet for Classic Wide Template adjustments
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.