Related Tables
The cytoskeleton is an essential component of a cell's structure and one of the easiest to label with fluorescent reagents. This section describes Invitrogen™ Molecular Probes™ labeling reagents for both monomeric actin (G-actin) and filamentous actin (F-actin); reagents for staining tubulin and other cytoskeletal proteins are described in Probes for Tubulin and Other Cytoskeletal Proteins—Section 11.2.
Fluorescent Actin
Alexa Fluor Actin and Unlabeled Actin
Fluorescently labeled actin (Figure 11.1.1) is an important tool for investigating the structural dynamics of the cytoskeleton.ref For example, the red-orange–fluorescent Alexa Fluor 568 conjugate of rabbit muscle actin has been injected into chick embryo fibroblasts, which were then fixed, permeabilized and stained with coumarin phallacidin (photo).
Fluorescent actin conjugates can be prepared by reacting amine residues of polymerized F-actin with the succinimidyl ester of the appropriate dye using a modification of the method described by Alberts and co-workers.ref After labeling, the conjugates are subjected to depolymerization and subsequent polymerization to help ensure that the actin conjugates are able to assemble properly. The labeled actin that polymerizes is then separated from remaining monomeric actin by centrifugation, depolymerized and stored in monomeric form.
probes-actin.par.43981.image.275.258.1.s001664-structure-uncomplexed-actin-gif
Figure 11.1.1 Ribbon diagram of the structure of uncomplexed actin in the ADP state. The four subdomains are represented in different colors, and ADP is bound at the center where the four subdomains meet. Four Ca2+ ions bound to the actin monomer are represented as gold spheres. In this structure, tetramethylrhodamine-5-maleimide (T6027) has been used to covalently attach the dye to a specific cysteine residue (Cys 374). Image provided by Roberto Dominguez, Boston Biomedical Research Institute, Watertown, Massachusetts. Reprinted with permission from Science (2001) 293:708. Copyright 2001 American Association for the Advancement of Science.
GFP- and RFP-Labeled Actin
The requirement for intracellular delivery of Alexa Fluor dye–labeled actin conjugates by microinjection typically limits their applications for live-cell imaging to experiments involving no more than a few (<10) cells. For applications such as high-content screening (HCS) assays requiring larger sample sizes, GFP–actin fusions are well-established probes for imaging cytoskeletal structure and dynamics.ref CellLight Actin-GFP (C10506C10582), CellLight Actin-RFP (C10502C10583Figure 11.1.2) expression vectors (CellLight reagents and their targeting sequences—Table 11.1) generate autofluorescent proteins fused to the N-terminus of human β-actin and incorporate all the generic advantages of BacMam delivery technology (BacMam Gene Delivery and Expression Technology—Note 11.1). In particular, the viral dose can be readily adjusted to modulate expression levels if GFP- or RFP-dependent perturbation of cellular structural or functional properties is a concern.
probes-actin.par.90080.image.275.204.1.s006947-actin-gif
Figure 11.1.2 HeLa cell labeled with CellLight Actin-RFP (C10502, C10583) and CellLight MAP4-GFP reagents and with Hoechst 33342 nucleic acid stain.
CellLight Null Control Reagent
The CellLight™ Null (control) reagent (C10615), a suspension of baculovirus particles lacking mammalian genetic elements, is designed for use in parallel with our CellLight™ targeted fluorescent protein reagents (CellLight reagents and their targeting sequences—Table 11.1). For example, microarray expression analysis on cells treated with the CellLight Null (control) reagent can be used to assess down-regulation or up-regulation of host cell genes elicited by baculovirus infection.
Phallotoxins for Labeling F-Actin
We prepare a number of fluorescent and biotinylated derivatives of phalloidin and phallacidin for selectively labeling F-actin. Phallotoxins are bicyclic peptides isolated from the deadly Amanita phalloides mushroom ref (www.grzyby.pl/gatunki/Amanita_phalloides.htm). They can be used interchangeably in most applications and bind competitively to the same sites on F-actin. Spectral characteristics of Molecular Probes actin-selective probes—Table 11.2 lists the available phallotoxin derivatives, along with their spectral properties.
A detailed staining protocol (Phallotoxins) is included with each phallotoxin derivative, and extensive product bibliographies are available at www.thermofisher.com. One vial of the fluorescent phallotoxin contains sufficient reagent for staining ~300 microscope slide preparations; one vial of biotin-XX phalloidin, which must be used at a higher concentration, contains sufficient reagent for ~50 microscope slide preparations. We also offer unlabeled phalloidin (P3457) for blocking F-actin staining by labeled phallotoxins and for promoting actin polymerization.
Properties of Phallotoxin Derivatives
The fluorescent and biotinylated phallotoxin derivatives stain F-actin selectively at nanomolar concentrations and are readily water soluble, thus providing convenient labels for identifying and quantitating actin in tissue sections, cell cultures or cell-free preparations.ref F-actin in live neurons can be efficiently labeled using cationic liposomes containing fluorescent phallotoxins, such as BODIPY™ FL phallacidin.ref This procedure permits the labeling of entire cell cultures with minimum disruption. Because fluorescent phalloidin conjugates are not permeant to most live cells, they can be used to detect cells that have compromised membranes. However, it has been reported that unlabeled phalloidin, and potentially dye-labeled phalloidins, can penetrate the membranes of certain hypoxic cells.ref An extensive study on visualizing the actin cytoskeleton with various fluorescent probes in cell preparations, as well as in live cells, has been published.ref
Labeled phallotoxins have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and nonmuscle cells; they reportedly do not bind to monomeric G-actin, unlike some antibodies against actin.ref Phallotoxins have further advantages over antibodies for actin labeling, in that 1) their binding properties do not change appreciably with actin from different species, including plants and animals; and 2) their nonspecific staining is negligible; thus, the contrast between stained and unstained areas is high.
Phallotoxins shift actin's monomer/polymer equilibrium toward the polymer, lowering the critical concentration for polymerization as much as 30-fold.ref Furthermore, depolymerization of F-actin by cytochalasins, potassium iodide and elevated temperatures is inhibited by phallotoxin binding. Because the phallotoxin derivatives are relatively small, with approximate diameters of 12–15 Å and molecular weights below 2000 daltons, a wide variety of actin-binding proteins—including myosin, tropomyosin, troponin and DNase I—can still bind to actin after treatment with fluorescent phallotoxins. Even more significantly, phallotoxin-labeled actin filaments retain certain functional characteristics; labeled glycerinated muscle fibers still contract, and labeled actin filaments still move on solid-phase myosin substrates.ref
Alexa Fluor Phalloidins
We have taken advantage of the outstanding fluorescence characteristics of our Alexa Fluor dyes (Alexa Fluor Dyes Spanning the Visible and Infrared Spectrum—Section 1.3) to create a series of Alexa Fluor dye–labeled phalloidins (photo, photo, photo, photo), which are widely used F-actin stains for many applications across the full spectral range. The Alexa Fluor phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability to other spectrally similar conjugates tested (photo). For improved fluorescence detection of F-actin in fixed and permeabilized cells, we encourage researchers to try these fluorescent phalloidins in their actin-labeling protocols. Alexa Fluor 647 phalloidin (A22287) and Alexa Fluor™ 660 phalloidin (A22285) are among the few probes available that can be excited by the 647 nm spectral line of the Ar-Kr laser. A series of videos showing Alexa Fluor 488 phalloidin–stained actin ref is available at the Journal of Cell Biology web site (www.jcb.org/cgi/content/full/150/2/361/DC1).
Oregon Green Phalloidins
Green-fluorescent actin stains are popular reagents for labeling F-actin in fixed and permeabilized cells. Unfortunately, the green-fluorescent fluorescein phalloidin and NBD phallacidin photobleach rapidly, making their photography difficult. We have our Oregon Green™ 488 dye (Fluorescein, Oregon Green and Rhodamine Green Dyes—Section 1.5) to prepare Oregon Green™ 488 phalloidin (O7466, photo). The excitation and emission spectra of the Oregon Green 488 dye are virtually superimposable on those of fluorescein, and Oregon Green 488 conjugates may be viewed with standard fluorescein optical filter sets. As shown in Figure 11.1.3, Oregon Green 514 phalloidin (available upon request, contact Custom Services for more information) is more photostable than fluorescein phalloidin, making it easier to visualize and photograph.
probes-actin.par.5146.image.275.236.1.s000213-photostability-gif
Figure 11.1.3 Photostability comparison for Oregon Green 514 phalloidin and fluorescein phalloidin (F432). CRE BAG 2 fibroblasts were fixed with formaldehyde, permeabilized with acetone and then stained with the fluorescent phallotoxins. Samples were continuously illuminated and images were acquired every 5 seconds using a Star 1 CCD camera (Photometrics); the average fluorescence intensity in the field of view was calculated with Image-1 software (Universal Imaging Corp.) and expressed as a fraction of the initial intensity. Three data sets, representing different fields of view, were averaged for each labeled phalloidin to obtain the plotted time courses.
BODIPY Phallotoxins
BODIPY FL and BODIPY™ 558/568 phallotoxins (B3475, photo) have some important advantages over the conventional NBD, fluorescein and rhodamine phallotoxins. BODIPY™ dyes are more photostable than these traditional fluorophores ref and have narrower emission bandwidths (BODIPY Dye Series—Section 1.4), making them especially useful for double- and triple-labeling experiments. BODIPY FL phallacidin, which reportedly gives a signal superior to that of fluorescein phalloidin,ref has been used for quantitating F-actin and determining its distribution in cells.ref The BODIPY FL phallacidin and BODIPY 558/568 phalloidin (B3475) exhibit excitation and emission spectra similar to those of fluorescein and rhodamine B, respectively, and can be used with standard optical filter sets.
Rhodamine Phalloidin and Other Red-Fluorescent Phalloidins
Rhodamine phalloidin (R415, photo) has been the standard for red-fluorescent phallotoxins.ref Rhodamine phalloidin is excited efficiently by the mercury-arc lamp in most fluorescence microscopes. However, our Alexa Fluor™ 546, Alexa Fluor 568, Alexa Fluor 594 and Texas Red™-X phalloidins ref (A22283, A12380, A12381, T7471; photo, photo) will be welcome replacements for rhodamine phalloidin in many multicolor applications because their emission spectra are better separated from those of the green-fluorescent Alexa Fluor 488, Oregon Green and fluorescein dyes.
Other Labeled Phallotoxins
The original yellow-green–fluorescent NBD phallacidin and green-fluorescent fluorescein phalloidin (F432) remain in use despite their relatively poor photostability (Figure 11.1.3). Photostability of fluorescein phalloidin and some other fluorescent phallotoxins can be considerably improved (photo) by mounting the stained samples with our ProLong™ Antifade Kit or ProLong™ Gold antifade reagent (P7481, P36930, P36934; Fluorescence Microscopy Accessories and Reference Standards—Section 23.1). We recommend the Alexa Fluor 488, Oregon Green 488 and BODIPY FL phallotoxins for photostable, green-fluorescent actin staining. Alexa Fluor™ 350 phalloidin (A22281) is the only blue-fluorescent phallotoxin conjugate currently available for staining actin.
Biotin-XX phalloidin (B7474) also permits detection of F-actin by electron microscopy and light microscopy techniques.ref This biotin conjugate can be visualized with fluorophore- or enzyme-labeled avidin and streptavidin (Avidin, Streptavidin, NeutrAvidin and CaptAvidin Biotin-Binding Proteins and Affinity Matrices—Section 7.6) or with tyramide signal amplification (TSA) technology (TSA and Other Peroxidase-Based Signal Amplification Techniques—Section 6.2). Biotin-XX phalloidin, in conjunction with streptavidin or CaptAvidin™ agarose (S951, C21386; Avidin, Streptavidin, NeutrAvidin and CaptAvidin Biotin-Binding Proteins and Affinity Matrices—Section 7.6), can be used to precipitate F-actin from the cytosolic anti-phosphotyrosine–reactive fraction in macrophages stimulated with colony-stimulating factor-1.ref
DNase I Conjugates for Staining G-Actin
Bovine pancreatic deoxyribonuclease (DNase I, ~31,000 daltons) binds much more strongly to monomeric G-actin than to filamentous F-actin, with binding constants of 5 × 108 M-1 and 1.2 × 104 M-1, respectively.ref Because of this strong, selective binding to G-actin, fluorescent DNase I conjugates have proven very useful for detecting and quantitating the proportion of unpolymerized actin in a cell. We have triple-labeled endothelial cells with fluorescein DNase I, BODIPY™ 581/591 phalloidin and a monoclonal anti-actin antibody detected with a Cascade Blue™ dye–labeled secondary antibody ref (C962, Secondary Immunoreagents—Section 7.2). We found that the monoclonal antibody, which binds to both G-actin and F-actin, colocalized with the DNase I and phalloidin conjugates, suggesting that these three probes recognize unique binding sites on the actin molecule. Researchers can choose DNase I conjugates labeled with either the green-fluorescent Alexa Fluor 488 (D12371) or red-fluorescent Alexa Fluor 594 (D12372) dyes, depending on their multicolor application and their detection instrumentation (Spectral characteristics of Molecular Probes actin-selective probes—Table 11.2).
Alexa Fluor 488 and Alexa Fluor 594 DNase I conjugates have been used in combination with fluorescently labeled phallotoxins to simultaneously visualize G-actin pools and filamentous F-actin ref and to study the disruption of microfilament organization in live nonmuscle cells.ref Rhodamine phalloidin (R415) has been used in conjunction with Oregon Green 488 DNase I to determine the F-actin:G-actin ratio in Dictyostelium using confocal laser-scanning microscopy.ref A mouse fibroblast labeled with both Texas Red DNase I and Oregon Green 488 phalloidin (O7466) permitted visualization of the G-actin and the complex network of F-actin throughout the cytoplasm, as well as at the cell periphery (photo). The influence of cytochalasins on actin structure in monocytes has been quantitated by flow cytometry using Texas Red DNase I and BODIPY FL phallacidin to stain the G-actin and F-actin pools, respectively.ref Fluorescent DNase I has also been used as a model system to study the interactions of nucleotides, cations and cytochalasin D with monomeric actin.ref
Probes for Actin Quantitation, Actin Polymerization and Actin-Binding Proteins
Assays for Quantitating F-Actin and G-Actin Polymerization
Quantitative assays for F-actin have employed fluorescein phalloidin,ref rhodamine phalloidin,ref BODIPY FL phallacidin ref and NBD phallacidin.ref An F-actin assay based on fluorescein phalloidin was used to demonstrate the loss of F-actin from cells during apoptosis.ref The addition of propidium iodide (P1304MP, P3566, P21493; Nucleic Acid Stains—Section 8.1) to the cell suspensions enabled these researchers to estimate the cell-cycle distributions of both the apoptotic and nonapoptotic cell populations. The change in F-actin content in proliferating adherent cells has been quantitated using the ratio of rhodamine phalloidin fluorescence to ethidium bromide fluorescence.ref The spectral separation of the signals in this assay may be improved by using a green-fluorescent stain for F-actin and a high-affinity red-fluorescent nucleic acid stain, such as the combination of Alexa Fluor 488 phalloidin (A12379) and ethidium homodimer-1 (E1169, Nucleic Acid Stains—Section 8.1).
The fluorescence of actin monomers labeled with pyrene iodoacetamide (P29) has been demonstrated to change upon polymerization, making this probe an excellent tool for following the kinetics of actin polymerization and the effects of actin-binding proteins on polymerization.ref
Jasplakinolide: A Cell-Permeant F-Actin Probe
We offer jasplakinolide (J7473), a macrocyclic peptide isolated from the marine sponge Jaspis johnstoni.ref Jasplakinolide is a potent inducer of actin polymerization in vitro by stimulating actin filament nucleation ref and competes with phalloidin for actin binding ref (Kd = 15 nM). Moreover, unlike other known actin stabilizers such as phalloidins and virotoxins, jasplakinolide appears to be somewhat cell permeant and therefore can potentially be used to manipulate actin polymerization in live cells. This peptide, which also exhibits fungicidal, insecticidal and antiproliferative activity,ref is particularly useful for investigating cell processes mediated by actin polymerization and depolymerization, including cell adhesion, locomotion, endocytosis and vesicle sorting and release. Jasplakinolide has been reported to enhance apoptosis induced by cytokine deprivation.ref
Latrunculin A and Latrunculin B: Cell-Permeant Actin Antagonists
Latrunculins are powerful disruptors of microfilament organization. Isolated from a Red Sea sponge, these G-actin binding compounds inhibit fertilization and early embryological development,ref alter the shape of cells ref and inhibit receptor-mediated endocytosis.ref Latrunculin A ref (L12370) binds to monomeric G-actin in a 1:1 ratio at submicromolar concentrations (Howard Petty, Wayne State University, personal communication) and is frequently used to establish the effects of F-actin disassembly on particular physiological functions such as ion transport ref and protein localization.ref The activity of latrunculin B mimics that of latrunculin A in most applications.ref
Assays for Actin-Binding Proteins
Enhancement of the fluorescence of certain phallotoxins upon binding to F-actin can be a useful tool for following the kinetics and extent of binding of specific actin-binding proteins. We have used the change in fluorescence of rhodamine phalloidin (R415) to determine the dissociation constant of various phallotoxins.ref The enhancement of rhodamine phalloidin's fluorescence upon actin binding has also been used to measure the kinetics and extent of gelsolin severing of actin filaments.ref The affinity and rate constants for rhodamine phalloidin binding to actin are not affected by saturation of actin with either myosin subfragment-1 or tropomyosin, indicating that these two actin-binding proteins do not bind to the same sites as the phalloidin.ref
Data Table
For a detailed explanation of column headings, see Definitions of Data Table Contents
Cat #MWStorageSolubleAbsECEmSolventNotes
A12379
Alexa Fluor 488 phalloidin
~1320F,LMeOH, H2O49478,000517pH 71, 2, 3
A12380
Alexa Fluor 568 phalloidin
~1590F,LMeOH, H2O57888,000600pH 71, 2, 3
A12381
Alexa Fluor 594 phalloidin
~1620F,LMeOH, H2O59392,000617pH 71, 2, 3
A22281
Alexa Fluor 350 phalloidin
~1100F,LMeOH, H2O34617,000446pH 71, 2, 3
A22282
Alexa Fluor 532 phalloidin
~1350F,LMeOH, H2O52881,000555pH 71, 2, 3
A22283
Alexa Fluor 546 phalloidin
~1800F,LMeOH, H2O554112,000570pH 71, 2, 3
A22284
Alexa Fluor 633 phalloidin
~1900F,LMeOH, H2O621159,000639MeOH1, 2, 3, 4
A22285
Alexa Fluor 660 phalloidin
~1650F,LMeOH, H2O668132,000697MeOH1, 2, 3, 4
A22286
Alexa Fluor 680 phalloidin
~1850F,LMeOH, H2O684183,000707MeOH1, 2, 3, 4
A22287
Alexa Fluor 647 phalloidin
~1950F,LMeOH, H2O650275,000672MeOH1, 2, 3, 4
A34054
Alexa Fluor 635 phalloidin
~1800F,LMeOH, H2O622145,000640MeOH1, 2, 3, 4
A34055
Alexa Fluor 555 phalloidin
~1900F,LMeOH, H2O555155,000572MeOH1, 2, 3
BODIPY FL phallacidin~1160F,LMeOH, H2O50583,000512MeOH1, 2, 3
B3475
BODIPY 558/568 phalloidin
~1115F,LMeOH, H2O55885,000569MeOH1, 2, 3
B7474
biotin-XX phalloidin
~1300FMeOH, H2O<300 none 1, 2
F432
fluorescein phalloidin
~1175F,LMeOH, H2O49684,000516pH 81, 2, 3
J7473
jasplakinolide
709.68F,DMeOH2788000noneMeOH 
L12370
latrunculin A
421.55F,DDMSO<300 none  
latrunculin B395.51F,DDMSO<300 none  
NBD phallacidin~1040F,LMeOH, H2O46524,000536MeOH1, 2, 3
Oregon Green 514 phalloidin~1280F,LMeOH, H2O51185,000528pH 91, 2, 3
O7466
Oregon Green 488 phalloidin
~1180F,LMeOH, H2O49686,000520pH 91, 2, 3
P29
N-(1-pyrene)iodoacetamide
385.20F,D,LDMF, DMSO33926,000384MeOH5, 6
P3457
phalloidin
~790FMeOH, H2O<300 see Notes 2, 7
R415
rhodamine phalloidin
~1250F,LMeOH, H2O54285,000565MeOH1, 2, 3, 8
T7471
Texas Red-X phalloidin
~1490F,LMeOH, H2O58395,000603MeOH1, 2, 3, 8
  1. α-Bungarotoxin, EGF and phallotoxin conjugates have approximately 1 label per peptide.
  2. Although this phallotoxin is water-soluble, storage in water is not recommended, particularly in dilute solution.
  3. The value of EC listed for this phallotoxin conjugate is for the labeling dye in free solution. Use of this value for the conjugate assumes a 1:1 dye:peptide labeling ratio and no change of EC due to dye–peptide interactions.
  4. In aqueous solutions (pH 7.0), Abs/Em = 625/645 nm for A22284, 633/648 nm for A34054, 649/666 nm for A22287, 661/689 nm for A22285 and 677/699 nm for A22286.
  5. Spectral data of the 2-mercaptoethanol adduct.
  6. Iodoacetamides in solution undergo rapid photodecomposition to unreactive products. Minimize exposure to light prior to reaction.
  7. This bicyclic peptide is very weakly fluorescent in aqueous solution (Em ~380 nm).ref
  8. In aqueous solutions (pH 7.0), Abs/Em = 554/573 nm for R415 and 591/608 nm for T7471.
Ordering Information
Catalog # Name Size Price (USD) Qty
A22281 Alexa Fluor™ 350 Phalloidin, Blue, 346/442 nm Each
640.65
Online Exclusive
660.00
Save 19.35 (3%)
A12379 Alexa Fluor™ 488 Phalloidin, Green, 495/518 nm Each
644.65
Online Exclusive
664.00
Save 19.35 (3%)
A22282 Alexa Fluor™ 532 Phalloidin, Yellow, 531/554 nm Each
615.65
Online Exclusive
634.00
Save 18.35 (3%)
A22283 Alexa Fluor™ 546 Phalloidin, Orange, 556/570 nm Each
642.65
Online Exclusive
662.00
Save 19.35 (3%)
A34055 Alexa Fluor™ 555 Phalloidin, Orange, 555/565 nm Each
659.65
Online Exclusive
678.00
Save 18.35 (3%)
A12380 Alexa Fluor™ 568 Phalloidin, Orange-red, 578/600 nm Each
628.65
Online Exclusive
660.00
Save 31.35 (5%)
A12381 Alexa Fluor™ 594 Phalloidin, Red, 581/609 nm Each
650.65
Online Exclusive
670.00
Save 19.35 (3%)
A22284 Alexa Fluor™ 633 Phalloidin, Far-red, 632/647 nm Each
647.65
Online Exclusive
668.00
Save 20.35 (3%)
A34054 Alexa Fluor™ 635 Phalloidin, Far-red, 633/647 nm Each
619.65
Online Exclusive
638.00
Save 18.35 (3%)
A22287 Alexa Fluor™ 647 Phalloidin, Far-red, 650/668 nm Each
644.65
Online Exclusive
668.00
Save 23.35 (3%)
A22285 Alexa Fluor™ 660 Phalloidin, Near-infrared, 663/690 nm Each
614.65
Online Exclusive
634.00
Save 19.35 (3%)
A22286 Alexa Fluor™ 680 Phalloidin, Near-infrared, 679/702 nm Each
628.65
Online Exclusive
656.00
Save 27.35 (4%)
B7474 Biotin-XX Phalloidin Each
611.65
Online Exclusive
630.00
Save 18.35 (3%)
B3475 BODIPY™ 558/568 Phalloidin, Red, 558/569 nm Each
598.65
Online Exclusive
616.00
Save 17.35 (3%)
C10582 CellLight™ Actin-GFP, BacMam 2.0, 1 mL Each
680.00
C10583 CellLight™ Actin-RFP, BacMam 2.0, 1 mL Each
648.65
Online Exclusive
672.00
Save 23.35 (3%)
C10615 CellLight™ Null (control), BacMam 2.0 Each -
D12371 Deoxyribonuclease I, Alexa Fluor™ 488 Conjugate Each
626.00
D12372 Deoxyribonuclease I, Alexa Fluor™ 594 Conjugate Each
602.00
F432 Fluorescein Phalloidin, Green, 496/516 nm Each
586.65
Online Exclusive
606.00
Save 19.35 (3%)
J7473 Jasplakinolide Each
655.65
Online Exclusive
682.00
Save 26.35 (4%)
L12370 Latrunculin A Each
636.65
Online Exclusive
656.00
Save 19.35 (3%)
O7466 Oregon Green™ 488 Phalloidin, Green, 496/520 nm Each
613.65
Online Exclusive
632.00
Save 18.35 (3%)
P3457 Phalloidin Each
624.65
Online Exclusive
642.00
Save 17.35 (3%)
R415 Rhodamine Phalloidin, Red-orange, 540/565 nm Each
605.65
Online Exclusive
624.00
Save 18.35 (3%)
T7471 Texas Red™-X Phalloidin, Red, 591/608 nm 300 units
609.65
Online Exclusive
628.00
Save 18.35 (3%)
For Research Use Only. Not for use in diagnostic procedures.