Nunc Cell Factory systems
When it comes to producing consistent quality vaccines and biologics designed to improve and save lives, nobody can do it alone.
With the same growth kinetics as laboratory-scale cell culture products, Nunc Cell Factory systems are a proven solution for large-scale production of cells, vaccines, and therapeutic proteins. Our hands-on expertise will help you accelerate productivity and ease the burden of regulatory compliance. This quality production cell culture platform reflects the rigorous standards that have made us the world's leading provider of cell culture products.
Contact us  Order now  Download brochure
Nunc Cell Factory systems are suitable for every step of your workflow
10-layer EasyFill Cell Factory system without assemblies
Nunc Cell Factory systems are a proven solution for research using adherent cell culture. These systems have the same growth kinetics as other laboratory-scale cell culture products.
Available in 1-layer to 40-layer formats, this solution enables consistent performance and enhanced productivity for scale-up.
Collection of Cell Factory accessories from tubing, connectors, ports, and related components
View our full line of accessories available for the Nunc Cell Factory systems, supporting the use of aseptic fluid handling methods.
Two Nunc Closed Cell Factory systems with tubing assemblies
The closed system Nunc Standard Cell Factory systems are built with standard components to help ensure quality as well as assurance of supply. This ready-to-use standard closed system helps you quickly develop and scale-up your adherent cell culture process.
Available in 2-layer and 10-layer formats, learn how you can get consistent, high-quality results with this ready-to-use closed system.
Array of Standard Cell Factory systems demonstrating the ability to configure in stacks of multiple plates from 1 to 40 layers
Achieve lot-to-lot consistency with a proven solution that helps you develop your process quickly. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling, and harvesting. Available in 1-layer to 40-layer formats.
Configure your Nunc Cell Factory system tubing assemblies easily
With true configured Nunc Cell Factory systems and equipment, advance your fully commercial-scale process with pre-assembled Cell Factory systems and automation equipment. Our commercial team will work with you to design a custom closed system and automation process that fit your specific needs.
Download interactive flyer
2, 10, and 40-layer Nunc High Density Cell Factory systems
Available in 1-layer to 40-layer formats, this solution is designed to optimize your process by increasing your output, while maintaining the same footprint as the standard Nunc Cell Factory systems.
Nunc Cell Factory manipulator with racks and carts
Realize process consistency, optimization, and standardization when scaling up multiple Nunc Cell Factory systems simultaneously with this trusted equipment.
10-layer EasyFill Cell Factory system without assemblies
Nunc Cell Factory systems are a proven solution for research using adherent cell culture. These systems have the same growth kinetics as other laboratory-scale cell culture products.
Available in 1-layer to 40-layer formats, this solution enables consistent performance and enhanced productivity for scale-up.
Collection of Cell Factory accessories from tubing, connectors, ports, and related components
View our full line of accessories available for the Nunc Cell Factory systems, supporting the use of aseptic fluid handling methods.
Two Nunc Closed Cell Factory systems with tubing assemblies
The closed system Nunc Standard Cell Factory systems are built with standard components to help ensure quality as well as assurance of supply. This ready-to-use standard closed system helps you quickly develop and scale-up your adherent cell culture process.
Available in 2-layer and 10-layer formats, learn how you can get consistent, high-quality results with this ready-to-use closed system.
Array of Standard Cell Factory systems demonstrating the ability to configure in stacks of multiple plates from 1 to 40 layers
Achieve lot-to-lot consistency with a proven solution that helps you develop your process quickly. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling, and harvesting. Available in 1-layer to 40-layer formats.
Configure your Nunc Cell Factory system tubing assemblies easily
With true configured Nunc Cell Factory systems and equipment, advance your fully commercial-scale process with pre-assembled Cell Factory systems and automation equipment. Our commercial team will work with you to design a custom closed system and automation process that fit your specific needs.
Download interactive flyer
2, 10, and 40-layer Nunc High Density Cell Factory systems
Available in 1-layer to 40-layer formats, this solution is designed to optimize your process by increasing your output, while maintaining the same footprint as the standard Nunc Cell Factory systems.
Nunc Cell Factory manipulator with racks and carts
Realize process consistency, optimization, and standardization when scaling up multiple Nunc Cell Factory systems simultaneously with this trusted equipment.
Nunc Cell Factory system and equipment video library
Nunc Standard Closed Call Factory System Overview
This video is an overview of the Thermo Scientific Nunc Standard Closed Cell Factory System. This video demonstrates a clean and closed system, designed to provide consistent and high-quality results to enhance your adherent cell culture processes.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextNunc Standard Closed Call Factory System Overview
          This video is an overview of the Thermo Scientific Nunc Standard Closed Cell Factory System. This video demonstrates a clean and closed system, designed to provide consistent and high-quality results to enhance your adherent cell culture processes.
        2. Now Playing
          Up NextHow To Fill and Empty Nunc Standard Cell Factory system
          Watch this step by step how to instructional video to learn how to fill and empty the Thermo Scientific Nunc Standard Cell Factory System, which is compatible with the same cell culture techniques used in smaller cell culture vessels.
        3. Now Playing
          Up NextHow To Fill and Empty Nunc Standard Closed Cell Factory system
          Watch this step by step how to instructional video to learn how to fill and empty the Thermo Scientific Nunc Standard Closed Cell Factory System, which is a closed system built with standard components to help ensure quality and assurance of supply.
        4. Now Playing
          Up NextThermo Scientific Nunc Cell Factory Incubator
          Mobile solution for the incubation of Nunc Cell Factory systems, available with and without CO2 control.
        5. Now Playing
          Up NextThermo Scientific Nunc Automatic Cell Factory Manipulator System (ACFM)
          Increase processing speed by eliminating manual filling, emptying, harvesting and trypsinization steps of all Cell Factory systems. Designed for large-scale cell culture and production of vaccines, cell therapeutics or other biological products.
        6. Now Playing
          Up NextThermo Scientific Nunc Cell Factory Shaker
          Automate the detachment of cells from Nunc Cell Factory Systems by using the Nunc Cell Factory Shaker System.
        7. Now Playing
          Up NextThermo Fisher Scientific single-use capacity expansion in Suzhou
          The Suzhou Manufacturing site produces the rigid containment, adherent cell culture products, BPCs and Fluid Transfer Assemblies. This video provides details on topics related to quality management systems, PPI implementation, product expansion, and
        8. Now Playing
          Up NextAseptic Cell Culture Scale Up Using Cell Factory System and BPC
          Connections made easy. Plug a media bag into any Cell Factory system for enhanced aseptic connectivity for filling or harvesting.
        9. Now Playing
          Up NextAseptic cell culture scale up using Nunc Cell Factory System, tubing set and peristaltic pump
          Ready. Set. Connect. See how aseptic filling is made easy when using a Nunc Cell Factory™ System, a PETG bottle with tubing set and a peristaltic pump.
        10. Now Playing
          Up NextHow to Use the Thermo Scientific Nunc EasyFill Cell Factory System
          Versatile and easy to use 'out of the box' system. Large, vented screw closure and versatile port design for pouring and aseptic filling.
      Related products
      Maximize productivity with automation solutions. Develop a fully commercial-scale process using Nunc Cell Factory equipment options.
      Let us help you customize your Nunc Cell Factory system to fit your specific needs. Use our guided checklist in this flyer to indicate desired layers, port size and locations, connector, vent filter, and tubing details. Get started now
      Ordering information
      Bioprocessing resources
      Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
      For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.