MicroAmp adhesive films can be used to quickly and ergonomically seal filled PCR plates to prevent sample evaporation and reduce the risk of sample cross contamination between wells. The adhesive films provide a secure seal over every well.
MicroAmp PCR accessories are convenient tools to facilitate PCR set up including bases/holders, capping, decapping, and adhesive sealing tools. 
MicroAmp Optical Adhesive Film
MicroAmp optical adhesive films are an easy-to-use film with a pressure-sensitive adhesive that is easy to handle with gloves and ensures a secure seal that does not interfere with real-time PCR readings. The GLPE (general purpose laboratory equipment) version of this seal is manufactured in an ISO registered facility making them suitable for use with diagnostic procedures. Most customers prefer the MicroAmp adhesive films listed in Table 1.
Table 1. Featured MicroAmp optical films and kits for qPCR.
MicroAmp Clear Adhesive Film
MicroAmp Clear Adhesive Film is recommended for general PCR applications. Simply peel the backing away from the adhesive and apply firmly across the top of the PCR plate. Sealed plates can be conveniently stacked without worrying about contamination.
Table 2. Featured MicroAmp adhesive films and accessories for thermal cycling.
MicroAmp PCR accessories
Increase your productivity with tools including bases to support PCR reaction tubes and 96-well reaction plates during reaction set up and post reaction processing. Support bases, AKA adaptor trays, help stabilize and increase readability of the tubes and plates to prevent errors. We offer a range of accessories to seal your tubes efficiently and effortlessly. Table 3 lists our popular accessories.
Table 3. Featured MicroAmp accessories for PCR and qPCR.
The PCR plastics Sustain Series are ISCC PLUS-certified, bio-based PCR plastics that help reduce lab’s carbon footprint. This represents our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance and are certified free of DNases, RNases, human DNA, and PCR inhibitors to help ensure the accuracy and reproducibility of PCR results.
Watch videos on how to use the MicroAmp accessories
How to seal a PCR plate
Video Player is loading.
Current Time 0:00
Duration 2:44
Loaded: 6.08%
Stream Type LIVE
Remaining Time 2:44
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    How to use adapter trays
    Video Player is loading.
    Current Time 0:00
    Duration 4:06
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 4:06
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      How to use PCR accessories
      Video Player is loading.
      Current Time 0:00
      Duration 4:10
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 4:10
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Education icon
         PCR enzymes education resources
        Access PCR learning resources—a knowledge base of all things PCR.
        Webinar icon
        Molecular biology webinars
        View our webinars for trends, tips, and tricks on molecular biology techniques.
        Video icon
        PCR plastics videos
        Watch videos on how to select and use the right consumables for your PCR and qPCR experiments.
        Tools icon
        Support center
        Find tips, troubleshooting help, and resources for your PCR and cDNA synthesis experiments.
        Computer mouse icon
        Contact us
        Email or call our Technical Application Scientists for questions regarding products for PCR workflow.
        Check icon
        OEM and custom PCR plastics
        Discover custom manufacturing solutions, product barcoding, and packaging capabilities tailored to your specifications.
        Stylesheet for Classic Wide Template adjustments