The Applied Biosystems VeritiPro Dx Thermal Cycler is an in vitro diagnostic (IVD) endpoint thermal cycler designed to amplify nucleic acids from human-derived specimens using polymerase chain reaction (PCR). It is manufactured to ISO 13485 and GMP requirements.
VeritiPro Dx Thermal Cycler conforms to:
  • EU 2017/746 requirements and is CE- IVD labeled in Europe
  • Classified as US-FDA class I medical device
  • UK Medical devices regulations 2002 and is UKCA labeled in UK
This instrument delivers precise temperature control with a ramp rate of 6.0°C/sec. There is no need for protocol re-optimization when upgrading from Veriti Dx Thermal Cycler; the simulation mode supports seamless transition.
The VeritiPro Dx 96-well Thermal Cycler is only available for sale in the United States, Europe, and other selected countries. Contact your nearest sales representative for more details.
Access virtual tour
Request a quote  View brochure
Key features of VeritiPro Dx Thermal Cyclers: In vitro diagnostic thermal cyclers
IVD mode for protocol security
The VeritiPro Dx Thermal Cycler features two security modes to provide user accessibility and safeguard the integrity of validated protocols and experimental results.
  • Creation mode to enable users to see and open all folders to modify, view, and run all methods
  • Run-only mode to restrict users to access the IVD folder, and only view and run IVD methods
Precise PCR optimization with VeriFlex block technology
Thermal block with VeriFlex technology
VeriFlex Blocks on the VeritiPro Dx Thermal Cycler help provide precise control of your PCR optimization. With six independent Peltier blocks, you can precisely set and control the temperature in each block, helping give you maximal versatility, flexibility, and confidence in your diagnostic development.
  • Run up to six precise and independent temperatures to determine the optimal annealing temperature for cycle sequencing or endpoint PCR
  • VeriFlex Blocks maintain their thermal characteristics between optimizing and isothermal conditions, minimizing the need for further optimization steps
Learn more about precise PCR optimization with VeriFlex Blocks
Easy programming with an intuitive touch screen
The powerful yet simple-to-operate user interface on the VeritiPro Dx system is driven with a large, 8-inch color TFT LCD touch screen for editing or creating new methods and real-time run monitoring. A convenient USB port is also built into the VeritiPro Dx to simplify data transfer and software updates.
VeritiPro Dx in vitro diagnostic thermal cycler services and support
scientist Emphasizing the need to ensure best service and support
Thermo Fisher Scientific offers a comprehensive range of services and support to help keep your VeritiPro Dx Thermal Cycler's performance at peak level. The following services are available at no additional cost upon instrument purchase:
Basic package for VeritiPro Dx Thermal Cyclers
  • On-site installation qualification and operational qualification (IQ/OQ) services at the time of instrument installation
  • Incidental post-repair IQ/OQ services
  • Standard 1-year factory warranty
Service contract package for VeritiPro Dx Thermal Cyclers
For labs operating in strictly regulated environments, the VeritiPro Dx Service Contract Package may be the ideal choice. The following service entitlements are additional to the basic package:
  • Extended 1-year factory warranty
  • Annual planned maintenance (PM) with OQ
The VeritiPro Dx Service Contract Package helps provide peace of mind with an extended warranty, annual testing, and documentation to help ensure that thermal cyclers in regulated environments have maximum uptime and produce high-quality data.
Qualification services for VeritiPro Dx Thermal Cyclers
Instrument hardware qualifications for VeritiPro Dx Thermal Cycler include IQ/OQ services to verify and document the instrument's ability to meet manufacturer design specifications. Thermo Fisher Scientific's qualification specialists can partner with the customer to help deliver timely, cost-effective, and trusted qualification services that include reliable and audit-style documentation to help ensure thermal cyclers meet regulatory requirements.
Specifications for the VeritiPro Dx Thermal Cycler
Block format96-well, 0.2 mL alloy block
Maximum block ramp rate6.0°C/sec
Temperature accuracy±0.25°C (35°C to 99.9°C)
Temperature range0°C to 100°C
Temperature uniformity<0.5°C (30 sec after reaching 95°C)
Reaction volume range10–100 µL
Veriflex block range6 VeriFlex zones
Supports: 25°C across block (5°C zone-to-zone) 
Display interface8 inch color TFT LCD touchscreen
Instrument memoryUSB, 8 GB on-board
Number of programsUp to 1,000 protocols
Dimensions (L x W x H)46.5 x 24.5 x 21.7 cm (18.3 in x 9.8 in x 8.5 in)
Weight12 kg (26.5 lbs)
Voltage100–120 V; 200–240 V; 50–60 Hz
Noise outputOperation <53 dB(A)
Idling <40 dB(A)
SustainabilitySleep (Standby mode) to reduce power consumption compared to idling mode
Ordering information
Catalog # Name Size Price (USD) Qty
4483348 MicroAmp™ EnduraPlate™ Optical 96-Well Clear GPLE Reaction Plates with Barcode, 20 Plates Each
219.65
Online exclusive
234.00
Save 14.35 (6%)
4481192 MicroAmp™ Optical 96-Well GPLE Reaction Plates with Barcode Each
112.00
4306311 MicroAmp™ Clear Adhesive Film Each
204.00
A49767 Optical Adhesive Covers GPLE Each
109.65
Online exclusive
115.00
Save 5.35 (5%)
4379983 MicroAmp™ 96-Well Tray for VeriFlex™ Blocks Each
98.65
Online exclusive
104.00
Save 5.35 (5%)
4381850 MicroAmp™ 96-Well Tray/Retainer Set for Veriti™ Systems Each
112.65
Online exclusive
119.00
Save 6.35 (5%)
A30588 MicroAmp™ Optical 8-Tube Strip with Attached Optical Caps, 0.2 mL Each
276.00
A30589 MicroAmp™ 8-Tube Strip with Attached Domed Caps, 0.2 mL Each
235.65
Online exclusive
247.00
Save 11.35 (5%)
N8010535 MicroAmp™ 8-Cap Strip, clear, 300 Strips Each
125.65
Online exclusive
137.00
Save 11.35 (8%)
N8010580 MicroAmp™ 8-Tube Strip, 0.2 mL Each
158.65
Online exclusive
168.00
Save 9.35 (6%)
N8010540 MicroAmp™ Reaction Tube with Cap, 0.2 mL, 1,000 tubes Each
177.00
N8010533 MicroAmp™ Reaction Tube without Cap, 0.2 mL, 2,000 tubes Each
340.00
Resources
PCR Solutions Brochure
PCR Thermal Cyclers Education
Plastics compatibility chart for Applied Biosystems instruments
PCR and cDNA Synthesis Support Center
Tips and troubleshooting help
School of Molecular Biology, PCR Education
Technical content for new and experienced molecular biologists
Thermal Cycler Software and Firmware Files
Download the latest updates for your instrument
Interested in private label thermal cyclers?
Submit an inquiry
Support
Service
We provide a full range of service options for all Applied Biosystems thermal cyclers.
View service plans
Instrument hardware qualification service
Have one of our engineers perform temperature qualifications at your site ›
*The VeritiPro Dx Thermal Cycler is available for in vitro diagnostic use in the United States of America and Europe and other selected countries only. The customer is responsible for any validation of assays, reagents, and consumables, and compliance with any regulatory requirements that pertain to their procedures and instrument use.
Stylesheet for Classic Wide Template adjustments