cell-therapy-banner-900x270
We have developed a trusted suite of Gibco Cell Therapy Systems (CTS) neural stem cell (NSC) culture media, supplements, substrates, and growth factors for cell therapy. These high quality GMP-manufactured products are designed to support your transition from discovery through clinical and commercial manufacturing, offering extensive safety testing and regulatory documentation.
Explore NSC media, supplements & growth factors for cell therapy
CTS Neurobasal Medium
Designed for maintenance and expansion of human embryonic or post-natal neural stem cells and primary neurons. Allows for pure populations of neuronal cells without the need for an astrocyte feeder layer.
CTS Neurobasal-A Medium
CTS Neurobasal-A Medium can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.
CTS B-27 Supplement XenoFree
Serum-free growth supplement that helps minimize safety risks associated with human-derived components for manufacturing of cell therapy products.
CTS N-2 Supplement
Serum-free, chemically defined supplement for the growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype.
CTS DPBS
CTS DPBS is the classic Dulbecco's phosphate-buffered saline, formulated without calcium, magnesium, and phenol red. It is manufactured in GMP, ISO-certified facilities to help ensure the quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, and the absence of bacterial/fungal contamination and endotoxin.
CTS Synth-a-Freeze Medium
Chemically defined, protein-free liquid cryopreservation medium intended for freezing and storing a variety of mammalian cell types including neural stem cells.
Gibco PeproTech growth factors and cytokines for neural stem cell therapy
product photo
Gibco PeproTech and PeproGMP growth factors and cytokines are available to support stem cell renewal, glial progenitor differentiation, basal and neuron-restricted progenitor differentiation, and more.
View available proteins
Discover closed, scalable cell processing and electroporation systems for your neural cell stem cell therapy research and manufacturing
CTS Rotea Counterflow Centrifugation System
  • Process flexibility
  • High cell recovery and viability
  • Low output volumes
  • Research through commercial manufacturing
Learn more
CTS Xenon Electroporation System
  • High speed and large volume
  • Scalable, proven performance and viability
  • Process flexibility
  • Efficient nonviral transfection
  • Closed-system processing
Learn more
Featured neural stem cell resources
Cell therapy resources
Support
Intended uses of the products mentioned on the page vary. For specific intended use statements, please refer to the product label.