Dionex Inuvion IC System: Ultra-reliable, day-to-day ion analysis
Perform ion analysis with the Thermo Scientific Dionex Inuvion IC System, an intuitive, ultra-reliable ion chromatography (IC) instrument with straightforward operation and extended application capabilities. This system supports electrolytic suppression and can include Reagent-Free IC (RFIC) eluent generation for additional time-savings, greater day-to-day consistency, and gradients to optimize methods. It offers automated start-up and shut-down routines to save time and consistently reliable performance for busy analytical testing laboratories.
Highlights
Flexible, adaptable platform
The Dionex Inuvion IC System is designed to help your lab meet the needs of today and the future. Upgrade the Dionex Inuvion IC System to include RFIC for automated eluent generation capabilities. Optional accessories can be swapped in and out of the instrument by users in less than five minutes for extended application capabilities.
Ultra-reliable performance
This state-of-the-art ion chromatography system is engineered for maximum productivity. With new pump technology and electronics, the Dionex Inuvion IC System provides enhanced day-to-day reliability. System self-diagnostics also detect any issues with hardware and consumables, giving you the confidence that it will be ready to run whenever you are.
Easy-to-use platform
The Dionex Inuvion IC System is designed for fast startup and maximum uptime. With convenient, unobstructed access to all necessary components and a logical flow path, users can quickly and safely access everything on the instrument.
Suppression compatibility
The Dionex Inuvion IC System offers the choice of using either chemical suppression, for more challenging samples, or electrolytic suppression, for ease of use and convenience. Suppressors remove conductive ions from the eluent to decrease background conductivity, while enhancing analyte signal, enabling high sensitivity to achieve low detection limits with conductivity detection.
High-pressure operation
With the ability to operate at up to 5,000 psi, the Dionex Inuvion IC System can take advantage of high-performance 4 µm-particle-size Thermo Scientific Dionex IC columns for more efficient separations that can be used to shorten run times.
Comprehensive service and support
Optimize the Dionex Inuvion IC system with the Unity™ Lab Services IonCare™ 5-year service plan*. This extensive plan includes coverage for parts, labor, and travel, and 2 preventive maintenance visits. Experience hassle-free, budget-friendly support that ensures your system functions smoothly and efficiently.
*IonCare service plan is available only in North America and Europe
Videos
System specifications
The table below provides an overview of the Dionex Inuvion IC System specifications. Not all specifications are listed. For a full list of the specifications, download the Dionex Inuvion IC System specifications sheet.
Specification
Dionex Inuvion IC System
Analytical pump and fluidics
Type
Serial dual-reciprocating pistons, microprocessor-controlled constant stroke, variable speed
Construction
Chemically inert, metal-free PEEK pump heads and flow paths compatible with aqueous eluents of pH 0–14 and reversed-phase solvents
Pump operating pressure
0–35 MPa (0–5000 psi)
Flow rate range
0.00–5.00 mL/min in 0.01 mL/min increments
Flow precision
<0.1%, typically
Flow accuracy
<0.1%, typically
Pressure ripple
<1%
Eluent on-off valve
Standard
Leak sensor
Optical, standard
Piston seal wash (optional)
Pump head wash can be operated in continuous or intermittent mode when connected to rinse solution supply
Pressure alarm limits
Upper and lower limit pressure alarms can be set
Vacuum degas
Standard, user adjustable vacuum level
Eluent bottles
Standard 2 L polypropylene bottle; allows various sizes
Eluent bottle pressure regulator (optional)
Supported by digital controlled regulator with display
Injection valve
6-port, 2-position Rheodyne valve, electronically activated
Columns supported
2, 3, 4, and 5 mm ID; maximum length 250 mm analytical column with 50 mm guard column
Eluent generator (RFIC configuration)
Eluent types
KOH, MSA, or K2CO3
Eluent concentration range
0.1-100 mM
Flow rates
0.10–3.00 mL/min when a Thermo Scientific Dionex EGC is installed
Maximum operating pressure
Dionex EGC cartridges: 35 MPa (5000 psi)
Gradient profiles
Up to 9 time-defined gradient steps
Column heater (optional)
Operating temperature range
10 to 60 °C (50 to 140 °F); settable within the software; minimum working range is 5 °C above ambient temperature
Temperature accuracy
±0.5 °C at sensor, at calibration points (35, 45 °C)
Suppressors and control
Chemical and electrolytic suppression
2 mm and 4 mm anion and cation suppressor types
Suppressor regeneration mode
Electrolytic suppressor – Recycle or external water mode
Chemical suppressor – External regenerant
Current control range
Thermo Scientific Dionex DRS 600 Electrolytically Regenerated Suppressor: 0–500 mA (4 mm) and 0–150 mA (2 mm) in 1 mA increments
Thermo Scientific Dionex ERD 500 Electrolytically Regenerated Desalter: 0–500 mA (4 mm) and 0–150 mA (2 mm) in 1 mA increments
Salt converter
Thermo Scientific Dionex SC-CERS 500 available in 2 and 4 mm versions
Carbonic acid removal for anions
Thermo Scientific Dionex CRD 200 Carbonate Removal Device for use with hydroxide eluents or Dionex CRD 300 for use with carbonate eluents
Non-suppressed chromatography
Yes, supported
Suppressor wear parts
Chemical suppression: optional regenerant pump
Electrolytic suppression: none
Dynamic suppression capacity
Anions:
  • Thermo Scientific Dionex ADRS 600 (4 mm): 200 μeq/min
  • Dionex ADRS 600 (2 mm): 50 μeq/min
  • Thermo Scientific Dionex AERS 500e (4 mm): 200 μeq/min
  • Dionex AERS 500e (2 mm): 50 μeq/min
  • Thermo Scientific Dionex AERS 500 Carbonate (4 mm): 30 μeq/min
  • Dionex AERS 500 Carbonate (2 mm): 7.5 μeq/min
  • Thermo Scientific Dionex ACRS 500 (4 mm): 150 μeq/min
  • Dionex ACRS 500 (2 mm): 75 μeq/min
Cations:
  • Thermo Scientific Dionex CDRS 600 (4 mm): 100 μeq/min
  • Dionex CDRS 600 (2 mm): 35 μeq/min
  • Thermo Scientific Dionex CERS 500e (4 mm): 100 μeq/min
  • Dionex CERS 500e (2 mm): 35 μeq/min
  • Thermo Scientific Dionex CCRS 500 (4 mm): 75 μeq/min
  • Dionex CCRS 500 (2 mm): 37.5 μeq/min
Void volumes
  • <50 μL for 4mm Dionex DRS 600, ERS 500e, ERS 500 Carbonate, and CRS 500 suppressors
  • <15 μL for 2mm Dionex DRS 600, ERS 500e, ERS 500 Carbonate, and CRS 500 suppressors
Conductivity detector electronics and flow cell
Type
Microprocessor-controlled digital signal processor
Cell drive
128 kHz square wave
Linearity
r2≥0.999%
Resolution
0.002 nS/cm
Full-scale output ranges
Digital input signal range 0-18,000 μS/cm, with auto-ranging; analog output signal range 0-18,000 μS/cm
Temperature compensation
Variable, default set at 1.7%/°C at cell temperature
Temperature range
Ambient +7 °C, 15 to 60 °C
Cell electrodes
Passivated 316 stainless steel; compatible with MSA
Cell body
Chemically inert polymeric material
Cell volume
<1 µL
Heat exchanger
Inert, tortuous path for low axial dispersion
Maximum cell operating pressure
10 MPa (1,500 psi)
Data filter
Rise times from 0 to 10 s, Data Collection Rate 1 to 100 Hz, user selectable
Autosampler
Automation using autosampler
For more information about available autosamplers, visit IC Autosamplers
System software
Chromeleon CDS, requires Microsoft® Windows® 10 or 11
Physical specifications
Dimensions (h × w × d)
66.1 x 29.2 x 43.2 cm (26.0 x 11.5 x 17.0 in.)
Weight
16.2 kg (36 lb)
Power requirements
100–240 V AC, 50–60 Hz autoranging
Operating temperature
4–40 °C (40–104 °F)
Operating humidity range
20-80% relative, non-condensing
Control modes
Full control through Chromeleon CDS software; alternative control through TTL or relay closures; one relay output, two TTL outputs, two assignable TTL inputs
USB communication protocol
One USB input; three USB outputs
Ordering information
Instruments
Optional accessories