UltraPure™ Tris Buffer (powder format)
UltraPure™ Tris Buffer (powder format)
Invitrogen™

UltraPure™ Tris Buffer (powder format)

UltraPure™ Tris Buffer is widely used because of its buffering range (pH 7.0 to 9.0) and compatibility with many enzymes,Read more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
15504020Promo Image1 kg
Catalog number 15504020
Price (USD)
161.65
Online Exclusive
175.00
Save 13.35 (8%)
Each
In stock
Add to cart
Quantity:
1 kg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
175.00
Save 13.35 (8%)
Each
Add to cart
Ask our AI about this Product
UltraPure™ Tris Buffer is widely used because of its buffering range (pH 7.0 to 9.0) and compatibility with many enzymes, including restriction endonucleases and DNA-modifying enzymes.

Performance and quality testing
No DNase, RNase, or protease activity detected.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialTris
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
FormPowder
Product LineUltraPure™
Product TypeTris Buffer
Quantity1 kg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit SizeEach
Contents & Storage
Contents: One container (1 kg) of UltraPure™ Tris buffering agent
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10170741Certificate of AnalysisApr 18, 202515504020
10170711Certificate of AnalysisJan 27, 202515504020
10170731Certificate of AnalysisDec 05, 202415504020
9724331Certificate of AnalysisFeb 09, 202415504020
9722381Certificate of AnalysisApr 11, 202315504020
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (4)

Citations & References
Abstract
MHC-II ubiquitination.
Authors:De Gassart A, De Angelis Rigotti F, Gatti E,
Journal:Methods Mol Biol
PubMed ID:23329511
'Ubiquitinated protein detection is often troublesome since in most cases this modification reduces the half-life of targeted proteins, inducing their degradation. Furthermore, ubiquitination is reversible thanks to the action of highly specific deubiquitinases present in all eukaryotic cells. MHC molecules ubiquitination has been demonstrated to be a key event in ... More
Pressure cycling technology (PCT) reduces effects of inhibitors of the PCR.
Authors:Marshall PL, King JL, Lawrence NP, Lazarev A, Gross VS, Budowle B,
Journal:Int J Legal Med
PubMed ID:22986474
A common problem in the analysis of forensic human DNA evidence, or for that matter any nucleic acid analysis, is the presence of contaminants or inhibitors. Contaminants may copurify with the DNA, inhibiting downstream PCR or they may present samples effectively as containing fewer templates than exist in the PCR, ... More
Expanding the diagnostic use of PCR in leptospirosis: improved method for DNA extraction from blood cultures.
Authors:Villumsen S, Pedersen R, Krogfelt KA, Jensen JS,
Journal:PLoS One
PubMed ID:20711446
Leptospirosis is a neglected zoonosis of ubiquitous distribution. Symptoms are often non-specific and may range from flu-like symptoms to multi-organ failure. Diagnosis can only be made by specific diagnostic tests like serology and PCR. In non-endemic countries, leptospirosis is often not suspected before antibiotic treatment has been initiated and consequently, ... More
Production of compartmented cultures of rat sympathetic neurons.
Authors:Campenot RB, Lund K, Mok SA,
Journal:Nat Protoc
PubMed ID:20010935
The compartmented culture, in which primary neurons plated in a proximal compartment send their axons under silicone grease barriers and into left and right distal compartments, has enhanced the experimental capabilities of neuronal cultures. Treatments can be applied separately to cell bodies/proximal axons or distal axons, and cell bodies/proximal axons ... More
4 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection