96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1
Thermo Scientific™

96 Well Plate, UpCell Temperature Responsive Surface, Pack of 1

Color:Clear; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberQuantity
1748978/Cs.
Catalog number 174897
Price (KRW)
771,000
Online offer
Ends: 30-Jun-2025
856,000
Save 85,000 (10%)
Case of 8
-
Add to cart
Quantity:
8/Cs.
Price (KRW)
771,000
Online offer
Ends: 30-Jun-2025
856,000
Save 85,000 (10%)
Case of 8
Add to cart
Ask our AI about this Product
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Microplates. Simply transfer the culture to room temperature to collect cells in suspension.
Thermo Scientific Nunc UpCell Surface
UpCell surface is a temperature-responsive cell culture surface that enables non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
A temperature-responsive poly(N-isopropylacrylamide) (PNIPAAm) polymer is grafted onto the standard cell culture treated polystyrene surface. At 37˚C, the covalently bound polymer allows cells to attach to the dish and grow as normal. At room temperature, the PNIPAAm binds water and swells, releasing the attached cells together with the extracellular matrices. The cells are dissociated from the culture substrate without trypsin.

Features:
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • Shelf Life: 3 years from date of manufacturing

Recommended for:
  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • For research use only
Specifications
Array8 x 12
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorClear
Description96F well plate, UpCell, clear, with lid, Sterile (Pack of 1, Case of 8)
No. per Case8
No. per Pack1
Packaging1/Pk., 8/Cs.
Product LineNunc™
Quantity8/Cs.
Surface TreatmentUpCell-Treated, Temperature Responsive
Well ShapeFlat
MaterialPolystyrene
No. of Wells96
SterilitySterile
TypeMicroplate
Volume (Metric) Working200 μL
Unit SizeCase of 8
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
NU96W2433Certificate of ComplianceDec 10, 2024174897
NU96W2312Certificate of ComplianceDec 06, 2024174897
NU96W2203Certificate of ComplianceDec 06, 2024174897
NU96W2022Certificate of ComplianceDec 06, 2024174897
NU96W2320Certificate of ComplianceDec 03, 2024174897
5 results displayed, search above for a specific certificate

Share catalog number, name or link