Pierce™ Anti-c-Myc Magnetic Beads
Pierce™ Anti-c-Myc Magnetic Beads
Thermo Scientific™

Pierce™ Anti-c-Myc Magnetic Beads

Perform immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
Have Questions?
Catalog NumberQuantity
888421 mL
Catalog number 88842
Price (USD)
454.65
Online Exclusive
506.00
Save 51.35 (10%)
Each
In stock
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
454.65
Online Exclusive
506.00
Save 51.35 (10%)
Each
Add to cart
Ask our AI about this Product

Thermo Scientific Pierce Anti-c-Myc Magnetic Beads are affinity particles for immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.

Features of Anti-c-Myc Magnetic Beads:

  • Specific—highly specific anti-c-Myc monoclonal antibody (clone 9E10) enables high yield and high purity immunoprecipitation
  • Convenient and fast—product instructions provide an easy-to-follow, optimized protocol for immunoprecipitation in approximately one hour
  • Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding for c-Myc-tag IP or co-IP experiments
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

The blocked magnetic bead surface is coated with anti-c-Myc antibody, a highly specific mouse IgG1 monoclonal antibody (clone 9E10) that recognizes the c-Myc-epitope tag (EQKLISEEDL) derived from the human c-myc oncogene (p62 c-myc). The Pierce Anti-c-Myc Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Specifications:

Product Details:

Pierce Anti-c-Myc Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant c-Myc tagged proteins and the co-immunoprecipitation (Co-IP) of their interacting proteins. The beads are incubated with a cell lysate containing c-Myc tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine (pH 2.0) 50 mM NaOH, or SDS-PAGE sample buffer, the protocol has been optimized for each of these conditions. Anti-c-Myc antibody can be used to detect c-Myc tagged protein by Western blot analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAntibody
Protein FormRecombinant
Quantity1 mL
Sufficient For100 μg GST-c-Myc (26kDa Fusion Protein) per mL of Bead Suspension
Targetc-Myc
Product LinePierce™
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Store at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3214725Certificate of AnalysisJun 03, 202588842
AC407212Certificate of AnalysisMar 26, 202588842
ZL403007Certificate of AnalysisDec 31, 202488842
ZI399900Certificate of AnalysisOct 15, 202488842
ZG395360Certificate of AnalysisAug 08, 202488842
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection