Silica Bead DNA Gel Extraction Kit
Silica Bead DNA Gel Extraction Kit
Thermo Scientific™

Silica Bead DNA Gel Extraction Kit

Thermo Scientific Silica Bead DNA Gel Extraction Kit is a simple and efficient system for DNA extraction from agarose gelsRead more
Have Questions?
Catalog NumberQuantity
K0513100 Preps
Catalog number K0513
Price (TWD)
5,950.00
Online offer
Ends: 30-Jun-2025
8,590.00
Save 2,640.00 (31%)
Each
-
Add to cart
Quantity:
100 Preps
Recurring order eligible. Learn more »
Price (TWD)
5,950.00
Online offer
Ends: 30-Jun-2025
8,590.00
Save 2,640.00 (31%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Silica Bead DNA Gel Extraction Kit is a simple and efficient system for DNA extraction from agarose gels and reaction mixtures. The kit utilizes the modified protocol of Vogelstein and Gillespie, employing solubilization of the agarose gel and selective adsorption of nucleic acids on specially prepared silica particles at high salt concentration. Silica particles bound with DNA are washed to remove contaminants and pure DNA is eluted with TE buffer or water.

This method requires few manipulations, and is both faster and easier to perform than other organic-based extraction methods. Compared to column based gel-extraction kits, the DNA Gel Extraction Kit offers flexibility for scaling reaction and elution volumes up or down. The purified DNA is suitable for all common molecular biology procedures, including restriction digestion, cloning, sequencing, etc.

Highlights

Flexible—easy to scale up or down both reaction and elution volumes
Efficient—greater than 80% DNA recovery
Fast—purification takes only 15 to 20 minutes
Effective—suitable for DNA fragments as short as 100 bp
Safe—does not involve phenol extraction

Applications

• Purification of DNA fragments from agarose gels prepared with TAE or TBE buffers
• Concentration of DNA samples for desalting or buffer change
• Removal of enzymes after PCR, restriction digestion, dephosphorylation or labeling reactions
• Removal of unincorporated nucleotides, primers and primer-dimers
• Removal of linkers following ligation
• Removal of residual salts, phenol, chloroform, or ethidium bromide
• Purification of RNA-free plasmid DNA

Includes Silica Powder Suspension Binding Buffer Concentrated Washing Buffer TBE Conversion Buffer Detailed Protocol

Note

Care should be taken to avoid DNA damage with UV light when the gel-purified DNA fragment will be used in downstream cloning reactions. Always use a long wavelength UV (360 nm) light-box during excision of the gel slice. If only a short-wavelength UV (254 to 312 nm) light-box is available, minimize the UV exposure to a few seconds or keep the gel on a glass or plastic plate. Alternatively, visible dyes can be included in standard agarose gels to allow for visualization of the DNA bands in ambient light (2, 3).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Nucleic Acid Labeling, Sequencing
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product TypeSilica Bead DNA Gel Extraction Kit
Quantity100 Preps
Sample TypeDNA, Gel Samples
TargetDNA from Enzymatic Reactions (e.g. PCR)
Test Time15 to 20 min.
FormatKit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3225071Certificate of AnalysisApr 24, 2025K0513
3225068Certificate of AnalysisApr 24, 2025K0513
3225054Certificate of AnalysisApr 24, 2025K0513
3224098Certificate of AnalysisApr 23, 2025K0513
3167172Certificate of AnalysisFeb 11, 2025K0513
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection