LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation
LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation
Invitrogen™

LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation

The LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit is used to determine the viability of cells prior to the fixationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
L34966400 Assays
L34957200 Assays
L3496580 Assays
Catalog number L34966
Price (USD)
423.00
Each
-
Add to cart
Quantity:
400 Assays
Recurring order eligible. Learn more »
Price (USD)
423.00
Each
Add to cart
Ask our AI about this Product
The LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation. This kit has been optimized and validated for use with a violet laser flow cytometer.

• Stable—dyes are freeze dried in separate vials to maintain stability

• Robust—staining pattern is the same before and after fixation

• Low compensation—minimal spectral overlap between other fluorophores

View a selection guide for all fixable viability dyes for flow cytometry.

Stable
Unlike products that are sold in solution, the LIVE/DEAD™ Fixable Aqua Stain has been conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time. Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated. If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles.

Robust
Dead cell discriminator stains can lose sensitivity after treatment with fixatives such as formaldehyde or ethanol-based fixation methods required for intracellular phosphorylation studies. The LIVE/DEAD™ Fixable Aqua Stain is an amine reactive dye that binds covalently to intracellular and extracellular amines, and the staining pattern is preserved following formaldehyde fixation.

Low compensation
The LIVE/DEAD™ Fixable Aqua Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser. The aqua-fluorescent reactive dye has an excitation maximum of ∼375 nm, but it is excited well with the 405 nm violet laser, and it has an emission maxima of ∼512 nm, so it can be collected in the second channel on most violet laser flow cytometers.

How it works
In cells with compromised membranes, the dye reacts with free amines both in the cell interior and on the cell surface, yielding intense fluorescent staining. In viable cells, the dye's reactivity is restricted to the cell-surface amines, resulting in less intense fluorescence. The difference in intensity is typically greater than 50-fold between live and dead cells, allowing for easy discrimination.

Colors available
LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet your multi-color panel needs.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityImpermeant
Cell TypeEukaryotic Cells
DescriptionLIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation
Detection MethodFluorescence
Dye TypeLIVE/DEAD™ Fixable Aqua Dead Cell Stain
FormSolid
FormatTube
Quantity400 Assays
Shipping ConditionRoom Temperature
ColorAqua
Emission526
Excitation Wavelength Range375 nm
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product LineLIVE/DEAD
Product TypeStain
Unit SizeEach
Contents & Storage
Contains 10 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Have questions about this product? Ask our AI assisted search.
What is the binding capacity of DynaGreen magnetic beads?
Can you comment on the non-specific binding when using DynaGreen magnetic beads to isolate exosomes?
Can I sonicate DynaGreen magnetic beads?
Can I freeze DynaGreen magnetic beads?
When using DynaGreen magnetic beads, how much should I dilute the lysate with RIPA buffer?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Which cell viability kits are compatible with fixation?

The LIVE/DEAD Fixable kits for flow cytometry analysis are compatible with fixation. These kits use amine-reactive cell-impermeant dyes that stain the cell surface of live cells and also the cytosol of dead cells-live cells are dim and dead cells are bright. Since the dye is covalently bound to the cells, it will be retained after fixation. Unfortunately, this method does not work well for imaging-based assays, as all cells are stained and it is difficult to distinguish bright dead cells from dim live cells with a microscope. Ethidium monoazide (EMA; Cat No. E1374) is a cell impermeant nucleic acid stain that can be applied to live cultures and stains only dead cells. After incubation and washing away unbound dye, the cells can be exposed to light to photoactivate EMA to crosslink to dead cell DNA. After crosslinking to dead cell DNA, the samples may be fixed and permeabilized. Image-IT DEAD Green Viability Stain (Cat. No. I10291) for imaging and high-content screening (HCS) analysis is a live-cell impermeant DNA binding dye that is compatible with fixation and permeabilization with good retention up to 48 hours. We also have a LIVE/DEAD Reduced Biohazard Cell Viability Kit (Cat. No. L7013) for imaging and flow analysis that contains two DNA binding dyes, SYTO 10 and Dead Red, that are sufficiently retained to be analyzed soon after 4% glutaraldehyde fixation.
Note: In general, DNA-binding dyes and calcein AM are not compatible with fixation, as these dyes are not covalently bound to components of the cell and will thus slowly diffuse out of cells after fixation, gradually staining all cells as dead.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Figures

Customers who viewed this item also viewed



Citations & References (5)

Citations & References
Abstract
Antibody to Langerin/CD207 localizes large numbers of CD8alpha+ dendritic cells to the marginal zone of mouse spleen.
Authors:Idoyaga J, Suda N, Suda K, Park CG, Steinman RM,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19168629
Dendritic cells (DCs) are strategically positioned to take up antigens and initiate adaptive immunity. One DC subset expresses CD8alphaalpha in mice and is specialized to capture dying cells and process antigens for MHC class I  ... More
Rapid up-regulation and granule-independent transport of perforin to the immunological synapse define a novel mechanism of antigen-specific CD8+ T cell cytotoxic activity.
Authors:Makedonas G, Banerjee PP, Pandey R, Hersperger AR, Sanborn KB, Hardy GA, Orange JS, Betts MR,
Journal:J Immunol
PubMed ID:19380804
CTL are endowed with the ability to eliminate pathogens through perforin-mediated cytotoxic activity. The mechanism for perforin-mediated Ag-specific killing has been solely attributed to cytotoxic granule exocytosis from activated CD8(+) T cells. In this study, we redefine this mechanism, demonstrating that virus-specific CD8(+) T cells rapidly up-regulate perforin in response ... More
?d T Cells Recognize a Microbial Encoded B Cell Antigen to Initiate a Rapid Antigen-Specific Interleukin-17 Response.
Authors:Zeng X, Wei YL, Huang J, Newell EW, Yu H, Kidd BA, Kuhns MS, Waters RW, Davis MM, Weaver CT, Chien YH,
Journal:Immunity
PubMed ID:22960222
?d T cells contribute uniquely to immune competence. Nevertheless, how they function remains an enigma. It is unclear what most ?d T cells recognize, what is required for them to mount an immune response, and how the ?d T cell response is integrated into host immune defense. Here, we report that a noted ... More
The microbial mimic poly IC induces durable and protective CD4+ T cell immunity together with a dendritic cell targeted vaccine.
Authors:Trumpfheller C, Caskey M, Nchinda G, Longhi MP, Mizenina O, Huang Y, Schlesinger SJ, Colonna M, Steinman RM,
Journal:Proc Natl Acad Sci U S A
PubMed ID:18256187
CD4(+) Th1 type immunity is implicated in resistance to global infectious diseases. To improve the efficacy of T cell immunity induced by human immunodeficiency virus (HIV) vaccines, we are developing a protein-based approach that directly harnesses the function of dendritic cells (DCs) in intact lymphoid tissues. Vaccine proteins are selectively ... More
Increased mucosal CD4+ T cell activation following vaccination with an adenoviral vector in rhesus macaques.
Authors:Bukh I, Calcedo R, Roy S, Carnathan DG, Grant R, Qin Q, Boyd S, Ratcliffe SJ, Veeder CL, Bellamy SL, Betts MR, Wilson JM,
Journal:
PubMed ID:24829340
The possibility that vaccination with adenoviral (AdV) vectors increased mucosal T-cell activation remains a central hypothesis to explain the potential enhancement of HIV acquisition within the Step trial. Modeling this within rhesus macaques is complicated because human adenoviruses, including adenovirus type 5 (HAdV-5), are not endogenous to macaques. Here, we ... More
5 total citations

Share catalog number, name or link

1x1 image pixel for data collection