Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry
Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry
Invitrogen™

Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry

Detect apoptosis via flow cytometry with Vybrant FLICA Caspase Assays that detect poly and caspase-3/7/8 activities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeDescription
V35118FAM, Hoechst 33342, Propidium IodideVybrant FAM Caspase-3 and -7 Assay Kit
V35117FMK, Hoechst 33342, Propidium IodideVybrant FAM Poly Caspases Assay Kit
V35119FAM, Hoechst 33344, Propidium IodideVybrant FAM Caspase-8 Assay Kit
Catalog number V35118
Price (USD)
346.00
25 assays
-
Add to cart
Label or Dye:
FAM, Hoechst 33342, Propidium Iodide
Description:
Vybrant FAM Caspase-3 and -7 Assay Kit
Recurring order eligible. Learn more »
Price (USD)
346.00
25 assays
Add to cart
Ask our AI about this Product
Quickly detect caspase-mediated apoptosis by flow cytometry with Vybrant FAM caspase assay kits for poly-caspases and caspases 3 and 7. The Vybrant FAM caspase assay kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity. Specifically, these caspase assay kits contain engineered FLICA substrates FAM-VAD-FMK (for poly-caspases), FAM-DEVD-FMK (for caspase-3/7), and FAM-LETD-FMK (for caspase-8). They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by flow cytometry.
The Vybrant FAM caspase assay kits for flow cytometry employ a novel approach to detecting active caspases: the kits take advantage of FLICA methodology, which is an affinity label. This affinity label associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase-3/7, and leucine-glutamic acid-threonine-aspartic acid (LETD) for caspase-8. A carboxyfluorescein (FAM) group is attached as a reporter.

The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488 nm/⁄530 nm, 535⁄ nm/6617 nm, and 350 nm/⁄461 nm, respectively. The detection reagents included within the individual Vybrant FAM Poly Caspases Assay, Caspase-3 and -7 Assay, and Caspase-8 Assay apoptosis kits are 488⁄⁄530 FAM-VAD-FMK reagent, 488/⁄530 FAM-DEVD-FMK, and 488/⁄530 FAM-LETD-FMK, respectively.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionVybrant FAM Caspase-3 and -7 Assay Kit
Excitation/EmissionFAM-DEVD-FMD: 388/530
PI: 535/617
Hoechst 33342: 350/461
Flow Cytometer Laser LinesUV, 488
For Use With (Equipment)Flow Cytometer
Kit Contents1 vial FAM-DEVD-FMK caspase-3 and -7 reagent (FLICA reagent, lyophilized solid), 1 vial of Hoechst 33342 (400 μL, 1 mM in water), 1 vial of PI (1 mL, 250 μg/mL in water), 1 vial of DMSO (0.5 mL), 1 bottle of apoptosis fixative solution (10% formaldehyde with methanol in PBS, 6 mL), and 1 bottle of 10x apoptosis wash buffer.
Label TypeOther Label(s) or Dye(s)
Label or DyeFAM, Hoechst 33342, Propidium Iodide
Product LineVybrant
Product TypeCaspase Assay Kit
Quantity25 assays
Shipping ConditionRoom Temperature
Storage RequirementsStore in refrigerator (2–8°C) and protect from light.
Detection MethodFluorescence
FormatTube
Unit Size25 assays
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3188405Certificate of AnalysisMay 04, 2025V35119
3136321Certificate of AnalysisApr 03, 2025V35119
3148244Certificate of AnalysisMar 30, 2025V35117
3004582Certificate of AnalysisDec 11, 2024V35119
2978397Certificate of AnalysisOct 25, 2024V35117
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

FAM-DEVD-FMK FLICA reagent: 488/530 nm
Hoechst 33342: 350/461 nm
Propidium iodide: 535/617 nm

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (43)

Citations & References
Abstract
Monocyte Chemoattractant Protein-Induced Protein 1 (MCPIP1) Enhances Angiogenic and Cardiomyogenic Potential of Murine Bone Marrow-Derived Mesenchymal Stem Cells.
Authors:Labedz-Maslowska A, Lipert B, Berdecka D, Kedracka-Krok S, Jankowska U, Kamycka E, Sekula M, Madeja Z, Dawn B, Jura J, Zuba-Surma EK,
Journal:
PubMed ID:26214508
'The current evidence suggests that beneficial effects of mesenchymal stem cells (MSCs) toward myocardial repair are largely due to paracrine actions of several factors. Although Monocyte chemoattractant protein-induced protein 1 (MCPIP1) is involved in the regulation of inflammatory response, apoptosis and angiogenesis, whether MCPIP1 plays any role in stem cell-induced ... More
Multiparameter detection of apoptosis using red-excitable SYTO probes.
Authors:Wlodkowic D, Skommer J, Hillier C, Darzynkiewicz Z,
Journal:Cytometry A
PubMed ID:18431792
'Functional assays allowing phenotypic characterization of different cell death parameters at a single-cell level are important tools for preclinical anticancer drug screening. Currently, the selection of cytometric assays is limited by the availability of fluorescent probes with overlapping spectral characteristics. Following on our earlier reports on green and orange fluorescent ... More
Mapping mechanisms and charting the time course of premature cell senescence and apoptosis: lysosomal dysfunction and ganglioside accumulation in endothelial cells.
Authors:Patschan S, Chen J, Gealekman O, Krupincza K, Wang M, Shu L, Shayman JA, Goligorsky MS,
Journal:Am J Physiol Renal Physiol
PubMed ID:17928415
'Endothelial cells subjected to glycated collagen I develop premature senescence within 3-5 days, as revealed by increased senescence-associated beta-galactosidase activity, decreased proliferation, and an increase in cell size. Here, we analyzed the time course and possible mechanisms of this process. Lysosomal integrity studies revealed a rapid collapse of pH gradient ... More
Activation of caspases measured in situ by binding of fluorochrome-labeled inhibitors of caspases (FLICA): correlation with DNA fragmentation.
Authors:Bedner E, Smolewski P, Amstad P, Darzynkiewicz Z
Journal:Exp Cell Res
PubMed ID:10942603
'Activation of caspases is the key event of apoptosis and new methods are needed to assay this event, particularly in situ, in individual cells. To measure in situ caspases activation in the present study we employed fam-VAD-fmk and fam-VEID-fmk, the fluorochrome (fam)-labeled inhibitors of caspases (FLICA), which through the fluoromethylketone ... More
Multiparametric evaluation of apoptosis: effects of standard cytotoxic agents and the cyanoguanidine CHS 828.
Authors:Lövborg H, Nygren P, Larsson R
Journal:Mol Cancer Ther
PubMed ID:15141009
'A multiparametric high-content screening assay for measurement of apoptosis was developed. HeLa cells and lymphoma U-937 cells were exposed to cytotoxic drugs in flat-bottomed optical microtiter plates. After incubation, the DNA-binding dye Hoechst 33342, fluorescein-tagged probes that covalently bind active caspases and chloromethyl-X-rosamine to detect mitochondrial membrane potential (MMP) were ... More
43 total citations

Share catalog number, name or link