Two-dimensional gel electrophoresis (2DE) is an established technique for high-resolution profiling of complex protein mixtures. 2DE can resolve thousands of protein “spots” on a single gel and is considered a key method in proteomics research. It is widely applied in protein expression profiling experiments to identify changes in protein expression levels resulting from a disease state or drug treatment. 2DE is also a valuable tool to study post-translational modifications and identify protein isoforms. Small changes in the protein mass or isoelectric point (pI) translate into a detectable protein shift.
2D electrophoresis consists of two separation steps. In the first-dimension, proteins are separated according to their isoelectric point (pI)—the pH at which the protein net charge is zero—in a process known as isoelectric focusing (IEF). During conventional IEF, amphoteric molecules, like proteins, migrate until they reach a region where the pH of the matrix matches their isoelectric point and they “focus” into sharp bands.
Novex IEF Gels can be used to determine the pI or to detect minor changes in a protein due to deamination, phosphorylation, or glycosylation. They are also excellent for native, nondenaturing applications using soluble proteins.
Immobilized pH gradient (IPG) represents an evolution of the technique. An immobilized pH gradient is produced in the gel before the sample is applied, resulting in improved resolution and reproducibility. The second dimension consists of a conventional SDS-PAGE electrophoretic separation where proteins are differentiated based on their molecular weights. 2-DE is a robust method in which the resolution acquired during the first dimension separation is preserved during the second dimension.
ZOOM IPGRunner System
Analyzing complex protein samples using traditional two-dimensional electrophoresis methods is tedious and time-consuming. The ZOOM IPGRunner System offers an integrated solution in a mini gel format. This innovative system makes sensitive protein profiling accessible to both novice and experienced proteomics researchers. The oil-free ZOOM IPGRunner System supports rapid experiment setup and execution and is a low-cost and time-saving solution for establishing the initial experimental conditions prior to running large-format 2D gel analysis.
You can obtain high-resolution results quickly and easily. The ZOOM IPGRunner System offers:
  • First dimension separation in as little as 3 hours
  • A unique design that eliminates messy mineral oil
  • Easy-to-handle IPG strips in wide and narrow one-unit pH ranges
  • Self-contained cassettes that simplify rehydration and focusing
  • High performance NuPage Bis-Tris gels provide a neutral pH environment for the second dimension that reduce in-gel protein degradation.
  • All components and reagents needed to get started with 2D electrophoresis are included in our first and second dimension welcome packs including PowerEase Touch HV Power Supply. Just add the IPG strips and ampholytes of your choice
The ZOOM IPGRunner System contains all the components necessary for easy, fast isoelectric focusing using IPG Strips
Sample preparation
Sample preparation is a fundamental step in any protein analysis workflow, but it is even more crucial in 2DE electrophoresis where no universal method exists. The buffer typically contains a denaturing agent, urea or mixture of urea and thiourea, a non-ionic or zwitterionic detergent, a reducing agent to cleave disulfide-bonds, and ampholytes. The solubilization buffer is formulated to achieve optimal protein solubilization and prevent precipitation during the rehydration of the IPG strips-supported gel. Salts, ionic molecules and ionic detergents interfere with isoelectric focusing and compromise protein separation. The table below lists reagents commonly used for sample preparation. The concentration of each component needs to be determined empirically and experimental conditions require optimization. This can be achieved easily and rapidly using the ZOOM IPGRunner System.
ComponentFunctionConcentration
Urea or Urea/Thiourea for membrane proteinsProtein denaturation and solubilization
8 M or 9 M urea or
5–8 M urea with 2 M thiourea
Non-ionic detergent (CHAPS, CHAPSO, NP-40)Protein solubilization and stabilization0.5–4%
DTT or DTEReduces disulfide bonds20 mM–100 mM
AmpholytesAid protein solubilization and maintain the pH gradient0.2–2%
First dimension
The ZOOM IPGRunner System is an oil-free platform for isoelectric focusing of up to 12 samples in as little as 3 hours. The 7 cm ZOOM Strips are immobilized pH gradient (IPG) gel strips cast on a plastic support. They are available in broad and medium pH range formats (3–10, 4–6, and 6–10).
IPG strips come in different pH ranges. They are available in broad linear and non-liner pH as well as in narrower pH ranges, expanding the resolution power of the system.
ZOOM IPG Strips options:
Broad range:Medium range:
pH 3–10 NonLinearpH 6–10
pH 3–10 LinearpH 4–7
Wide pH range IPG strips allow the separation of most proteins in a single gel. The non-linear pH gradient improves the resolution of proteins in the middle of the pH range, where most proteins tend to focus, by expanding the pH range in the middle of the gel and compressing the pH at the acidic and basic ends of the gradient. By running IPG strips with overlapping pH range, the overall resolution of the separation is improved and more proteins can be analyzed, without a time-consuming sample fractionation step that can affect reproducibility.
The table below shows protein loading recommendation.
IPG pH RangeCoomassie StainingSilver StainingVolume
Broad (3–10 L, NL)20–50 μg5–15 μg140 μl or 155 μl for overnight rehydration
Medium (6–10, 4–7)100–200 μg50–100 μg140 μl or 155 μl for overnight rehydration
The ZOOM Carrier Ampholytes are small, soluble molecules with both positive and negative charge groups that sort at their relative position based upon their isoelectric field, setting up a pH gradient. Carrier ampholytes help to stabilize the pH gradient and current in IPG strips and aid in protein solubility, resulting in more reproducible IEF resolution.
Second dimension
Following isoelectric focusing, the ZOOM Strips are reduced, alkylated and equilibrated with NuPAGE LDS Sample Buffer. Equilibrated ZOOM IPG strips load easily into the NuPAGE Gels for 2D separation of proteins by molecular weight.
Protein gel staining
Following the SDS-PAGE, separated protein spots can be stained and visualized. The following protein stains combine high sensitivity with rapid and easy staining protocols and are compatible with downstream excision of the protein spot, tryptic-digestion and mass-spectrometry analysis.
Protein Stain SensitivityStaining timeAdditional Information
SYPRO Ruby0.25–1 ng90 min microwave
18 hr standard
Fluorescent stain           
Ex: 280, 450nm
Em: 610 nm
SilverQuest Silver Staining Kit0.3 ngStd: 1 hr 30 min
Microwave: 30 min
Includes ingredients for
complete removal of
silver ions 
Simply Blue Safe Stain> 7 ng12 min microwave    
135 min standard
Single component–
Ready to use
Pro-Q Emerald 488 Glycoprotein Gel and Blot Stain kit4 ng~6 hrEx: 510 nm
Em: 520 nm
Pro-Q Emerald 300 Glycoprotein Gel and Blot Stain kit0.5 ng~5 hrEx: 280 nm
Em: 530 nm
Can also be detected with 300 nm UV illumination
Pro-Q Diamond Phosphoprotein Gel Staining Kit1-16 ng4–5 hrEx: 555 nm
Em: 580 nm
Protein spots can also be transferred to a membrane for immunodetection or for protein sequencing.
Welcome packs
Our first and second dimension welcome packs combine all the components necessary to get started with 2D electrophoresis. Just add the ZOOM IPG strips and carrier ampholytes that suit your needs.
See the table below for a complete list of the products included in each welcome pack:
 NameComponents
First dimensionPowerEase™ Touch HV Power Supply and ZOOM™ IPGRunner™ Welcome PackPE Touch HV
ZOOM IPGRunner Mini-Cell
ZOOM IPGRunner Cassettes
ZOOM Urea
ZOOM CHAPS
ZOOM Equilibration Trays
Second dimensionNuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%, IPG-WellXCell SureLock Mini-Cell
NuPAGE 4–12% Bis-Tris ZOOM gels
NuPAGE LDS Sample Buffer (4X)
NuPAGE Sample Reducing Agent (10X)
NuPage MES Running Buffer (20X)
PageRuler Plus Prestained Protein Ladder
Ordering
First dimension
Catalog # Name Size Price (USD) Qty
PSHV00A PowerEase™ Touch HV Power Supply Adapters Each
110.00
PSHV00 PowerEase™ Touch HV Power Supply, 115 VAC Each
3,929.65
Online Exclusive
4,115.00
Save 185.35 (5%)
ZM0003 ZOOM™ IPGRunner™ Cassette 10 cassette(s)
544.65
Online Exclusive
562.00
Save 17.35 (3%)
ZM0025 ZOOM™ Carrier Ampholytes 4-6, 10mL 10 mL
410.00
ZM0002 ZOOM™ IPGRunner™ Combo Kit 1 kit
2,031.65
Online Exclusive
2,125.00
Save 93.35 (4%)
ZM0013 ZOOM™ IPG Strip, pH 6-10 12 strips
195.65
Online Exclusive
201.00
Save 5.35 (3%)
ZM0024 ZOOM™ Carrier Ampholytes pH 9-11 10 mL
415.00
ZM0001 ZOOM™ IPGRunner™ Mini-cell 1 system
1,652.00
ZM0012 ZOOM™ IPG Strip, pH 4-7 12 strips
201.00
ZM0023 ZOOM™ Carrier Ampholytes pH 6-9 10 mL
421.00
ZM0011 ZOOM™ IPG Strips, pH 3-10NL (non-linear) 12 strips
201.00
ZM0022 ZOOM™ Carrier Ampholytes pH 4-7 10 mL
421.00
ZM0021 ZOOM™ Carrier Ampholytes pH 3-10 10 mL
418.00
PSHV00Z PowerEase™ Touch HV Power Supply and ZOOM™ IPGRunner Welcome Pack Each
6,110.00
ZM0008 ZOOM™ IPGRunner™ Cassettes and Trays 10 trays, 10 cassette(s)
696.00
ZM0007 ZOOM™ Equilibration Trays 10⁄pack
120.65
Online Exclusive
124.00
Save 3.35 (3%)
ZM0018 ZOOM™ IPG Strips, pH 3-10L 12 strips
195.65
Online Exclusive
201.00
Save 5.35 (3%)
Second dimension
Catalog # Name Size Price (USD) Qty
NP0330BOX NuPAGE™ 4 to 12%, Bis-Tris ZOOM™, 1.0 mm, Mini Protein Gel, IPG-well, 10 gels (1 box) Each
195.65
Online Exclusive
204.00
Save 8.35 (4%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
80.65
Online Exclusive
84.75
Save 4.10 (5%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
185.00
NP0330Z NuPAGE™ Bis-Tris IPG-well Welcome Pack Each
984.00
Protein gel staining
Catalog # Name Size Price (USD) Qty
S12000 SYPRO™ Ruby Protein Gel Stain, 1 L Each
445.65
Online Exclusive
465.00
Save 19.35 (4%)
P21857 Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit Each
393.00
LC6070 SilverQuest™ Silver Staining Kit Each
448.65
Online Exclusive
468.00
Save 19.35 (4%)
LC6065 SimplyBlue™ SafeStain, 3.5 L Each
550.00
P33300 Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L Each
649.65
Online Exclusive
668.00
Save 18.35 (3%)
P21875 Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit Each
368.00
Sample preparation
Sample preparation is a fundamental step in any protein analysis workflow, but it is even more crucial in 2DE electrophoresis where no universal method exists. The buffer typically contains a denaturing agent, urea or mixture of urea and thiourea, a non-ionic or zwitterionic detergent, a reducing agent to cleave disulfide-bonds, and ampholytes. The solubilization buffer is formulated to achieve optimal protein solubilization and prevent precipitation during the rehydration of the IPG strips-supported gel. Salts, ionic molecules and ionic detergents interfere with isoelectric focusing and compromise protein separation. The table below lists reagents commonly used for sample preparation. The concentration of each component needs to be determined empirically and experimental conditions require optimization. This can be achieved easily and rapidly using the ZOOM IPGRunner System.
ComponentFunctionConcentration
Urea or Urea/Thiourea for membrane proteinsProtein denaturation and solubilization
8 M or 9 M urea or
5–8 M urea with 2 M thiourea
Non-ionic detergent (CHAPS, CHAPSO, NP-40)Protein solubilization and stabilization0.5–4%
DTT or DTEReduces disulfide bonds20 mM–100 mM
AmpholytesAid protein solubilization and maintain the pH gradient0.2–2%
First dimension
The ZOOM IPGRunner System is an oil-free platform for isoelectric focusing of up to 12 samples in as little as 3 hours. The 7 cm ZOOM Strips are immobilized pH gradient (IPG) gel strips cast on a plastic support. They are available in broad and medium pH range formats (3–10, 4–6, and 6–10).
IPG strips come in different pH ranges. They are available in broad linear and non-liner pH as well as in narrower pH ranges, expanding the resolution power of the system.
ZOOM IPG Strips options:
Broad range:Medium range:
pH 3–10 NonLinearpH 6–10
pH 3–10 LinearpH 4–7
Wide pH range IPG strips allow the separation of most proteins in a single gel. The non-linear pH gradient improves the resolution of proteins in the middle of the pH range, where most proteins tend to focus, by expanding the pH range in the middle of the gel and compressing the pH at the acidic and basic ends of the gradient. By running IPG strips with overlapping pH range, the overall resolution of the separation is improved and more proteins can be analyzed, without a time-consuming sample fractionation step that can affect reproducibility.
The table below shows protein loading recommendation.
IPG pH RangeCoomassie StainingSilver StainingVolume
Broad (3–10 L, NL)20–50 μg5–15 μg140 μl or 155 μl for overnight rehydration
Medium (6–10, 4–7)100–200 μg50–100 μg140 μl or 155 μl for overnight rehydration
The ZOOM Carrier Ampholytes are small, soluble molecules with both positive and negative charge groups that sort at their relative position based upon their isoelectric field, setting up a pH gradient. Carrier ampholytes help to stabilize the pH gradient and current in IPG strips and aid in protein solubility, resulting in more reproducible IEF resolution.
Second dimension
Following isoelectric focusing, the ZOOM Strips are reduced, alkylated and equilibrated with NuPAGE LDS Sample Buffer. Equilibrated ZOOM IPG strips load easily into the NuPAGE Gels for 2D separation of proteins by molecular weight.
Protein gel staining
Following the SDS-PAGE, separated protein spots can be stained and visualized. The following protein stains combine high sensitivity with rapid and easy staining protocols and are compatible with downstream excision of the protein spot, tryptic-digestion and mass-spectrometry analysis.
Protein Stain SensitivityStaining timeAdditional Information
SYPRO Ruby0.25–1 ng90 min microwave
18 hr standard
Fluorescent stain           
Ex: 280, 450nm
Em: 610 nm
SilverQuest Silver Staining Kit0.3 ngStd: 1 hr 30 min
Microwave: 30 min
Includes ingredients for
complete removal of
silver ions 
Simply Blue Safe Stain> 7 ng12 min microwave    
135 min standard
Single component–
Ready to use
Pro-Q Emerald 488 Glycoprotein Gel and Blot Stain kit4 ng~6 hrEx: 510 nm
Em: 520 nm
Pro-Q Emerald 300 Glycoprotein Gel and Blot Stain kit0.5 ng~5 hrEx: 280 nm
Em: 530 nm
Can also be detected with 300 nm UV illumination
Pro-Q Diamond Phosphoprotein Gel Staining Kit1-16 ng4–5 hrEx: 555 nm
Em: 580 nm
Protein spots can also be transferred to a membrane for immunodetection or for protein sequencing.
Welcome packs
Our first and second dimension welcome packs combine all the components necessary to get started with 2D electrophoresis. Just add the ZOOM IPG strips and carrier ampholytes that suit your needs.
See the table below for a complete list of the products included in each welcome pack:
 NameComponents
First dimensionPowerEase™ Touch HV Power Supply and ZOOM™ IPGRunner™ Welcome PackPE Touch HV
ZOOM IPGRunner Mini-Cell
ZOOM IPGRunner Cassettes
ZOOM Urea
ZOOM CHAPS
ZOOM Equilibration Trays
Second dimensionNuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%, IPG-WellXCell SureLock Mini-Cell
NuPAGE 4–12% Bis-Tris ZOOM gels
NuPAGE LDS Sample Buffer (4X)
NuPAGE Sample Reducing Agent (10X)
NuPage MES Running Buffer (20X)
PageRuler Plus Prestained Protein Ladder
Ordering
First dimension
Catalog # Name Size Price (USD) Qty
PSHV00A PowerEase™ Touch HV Power Supply Adapters Each
110.00
PSHV00 PowerEase™ Touch HV Power Supply, 115 VAC Each
3,929.65
Online Exclusive
4,115.00
Save 185.35 (5%)
ZM0003 ZOOM™ IPGRunner™ Cassette 10 cassette(s)
544.65
Online Exclusive
562.00
Save 17.35 (3%)
ZM0025 ZOOM™ Carrier Ampholytes 4-6, 10mL 10 mL
410.00
ZM0002 ZOOM™ IPGRunner™ Combo Kit 1 kit
2,031.65
Online Exclusive
2,125.00
Save 93.35 (4%)
ZM0013 ZOOM™ IPG Strip, pH 6-10 12 strips
195.65
Online Exclusive
201.00
Save 5.35 (3%)
ZM0024 ZOOM™ Carrier Ampholytes pH 9-11 10 mL
415.00
ZM0001 ZOOM™ IPGRunner™ Mini-cell 1 system
1,652.00
ZM0012 ZOOM™ IPG Strip, pH 4-7 12 strips
201.00
ZM0023 ZOOM™ Carrier Ampholytes pH 6-9 10 mL
421.00
ZM0011 ZOOM™ IPG Strips, pH 3-10NL (non-linear) 12 strips
201.00
ZM0022 ZOOM™ Carrier Ampholytes pH 4-7 10 mL
421.00
ZM0021 ZOOM™ Carrier Ampholytes pH 3-10 10 mL
418.00
PSHV00Z PowerEase™ Touch HV Power Supply and ZOOM™ IPGRunner Welcome Pack Each
6,110.00
ZM0008 ZOOM™ IPGRunner™ Cassettes and Trays 10 trays, 10 cassette(s)
696.00
ZM0007 ZOOM™ Equilibration Trays 10⁄pack
120.65
Online Exclusive
124.00
Save 3.35 (3%)
ZM0018 ZOOM™ IPG Strips, pH 3-10L 12 strips
195.65
Online Exclusive
201.00
Save 5.35 (3%)
Second dimension
Catalog # Name Size Price (USD) Qty
NP0330BOX NuPAGE™ 4 to 12%, Bis-Tris ZOOM™, 1.0 mm, Mini Protein Gel, IPG-well, 10 gels (1 box) Each
195.65
Online Exclusive
204.00
Save 8.35 (4%)
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
80.65
Online Exclusive
84.75
Save 4.10 (5%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
185.00
NP0330Z NuPAGE™ Bis-Tris IPG-well Welcome Pack Each
984.00
Protein gel staining
Catalog # Name Size Price (USD) Qty
S12000 SYPRO™ Ruby Protein Gel Stain, 1 L Each
445.65
Online Exclusive
465.00
Save 19.35 (4%)
P21857 Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit Each
393.00
LC6070 SilverQuest™ Silver Staining Kit Each
448.65
Online Exclusive
468.00
Save 19.35 (4%)
LC6065 SimplyBlue™ SafeStain, 3.5 L Each
550.00
P33300 Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L Each
649.65
Online Exclusive
668.00
Save 18.35 (3%)
P21875 Pro-Q™ Emerald 488 Glycoprotein Gel and Blot Stain Kit Each
368.00
References
  1. Boldt, A, Fortunato, D, Conti, A, Petersen, A, Ballmer-Weber, B, Lepp, U, Reese, G, and Becker, W-M “Analysis of the composition of an immunoglobulin E reactive high molecular weight protein complex of peanut extract containing Ara h 1 and Ara h 3/4” Proteomics 2005 Feb;5(3):675-86.
  2. Tetlow, IJ, Wait, R, Lu, Z, Akkasaeng, R, Bowsher, CG, Esposito, S, Kosar-Hashemi, B, Morell, MK, and Emes, MJ “Protein Phosphorylation in Amyloplasts Regulates Starch Branching Enzyme Activity and Protein-Protein Interactions” Plant Cell 2004 Mar;16:694-708.
  3. Zancai P, Dal Col J, Piccinin S, Guidoboni M, Cariati R, Rizzo S, Boiocchi M, Maestro R, Dolcetti R. “Retinoic acid stabilizes p27(Kip1) in EBV-immortalized lymphoblastoid B cell lines through enhanced proteasome-dependent degradation of the p45(Skp2) and Cks1 proteins” Oncogene. 2005 Feb 14; 
  4. O’Farrell PH: High resolution two-dimensional electrophoresis of proteins. J Biol Chem. 1975, 250: 4007-4021.
  5. Righetti, P. G. Isoelectric Focusing: Theory, Methodology and Applications. Elsevier Biomedical, Amsterdam. 1983
  6. Righetti, P. G. Immobilized pH Gradients: Theory and Methodology. Elsevier, Amsterdam. 1990
For Research Use Only. Not for use in diagnostic procedures.