Vanquish Loop Heart-Cut 2D-LC Systems
Confirm peak purity for more confidence in complex analysis.
Vanquish Loop Heart-Cut 2D-LC Systems
Increase your confidence in peak purity using the Thermo Scientific Vanquish Loop Heart-Cut 2D-LC System. This two-dimensional LC system (LC-LC) enables you to collect one or multiple fractions of the first-dimension separation in a sample loop and transfer the cut-out fraction(s) in an automated workflow to a second-dimension separation. Heart-Cut 2D-LC can reveal hidden impurities that co-elute with the analytes of interest by using two complementary column chemistries to gain more separation power. The system is configured using modules from the Vanquish Core HPLC, Vanquish Flex UHPLC or Horizon UHPLC systems, retaining all the performance, robustness and ease of use of the Vanquish LC platform.
Highlights
Greater confidence
Collect one or multiple fractions of the first-dimension separation in a sample loop and transfer the cut-out fraction(s) to a second-dimension column with complementary column chemistry in an automated workflow for more confidence in peak purity.
Flexibility 
The modularity of Vanquish LC systems makes it easy to build a system to best suit your application requirements and upgrade a standard HPLC instrument to more advanced 2D-LC configurations when you need to address more challenging analyses.
Save time and resources
The integrated LC-LC system in combination with automated workflows, with predefined fractionation and transfer steps, allow for unattended sample analysis while reducing human error and saving time and bench space compared to offline approaches.
Easy and reliable tool-free connections
The Vanquish Loop Heart-Cut 2D-LC system is fitted with Thermo Scientific Viper fingertight fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup
CDS software
The Vanquish 2D-LC systems seamlessly integrate with our Thermo Scientific Chromeleon Chromatography Data System (CDS) to deliver superior compliance tools, networking capabilities, automation and data processing. 
What is loop-based heart-cut 2D-LC?
Our loop-based heart-cut 2D-LC system allows you to automate and cut-out fractions(s) of the eluent of the first-dimension (¹D) column into a sample loop to undergo a second separation on a complementary second-dimension (²D) column to resolve difficult or co-eluting peaks.
This flow scheme illustrates a Vanquish Online 2D-LC system for multiple heart-cutting by storing the cut-out fractions in multiple loops before performing a complementary analysis on a second-dimension (²D) column.
Flow schemes of the different steps in multi-heart-cut 2D-LC (example for loop 1)
Videos
System specifications
The table below provides an overview of the Vanquish Loop Heart-Cut 2D-LC system specifications for standard setups. Not all specifications are listed. Specifications may differ based on the specific system components selected. See detailed Component Specifications for more information.
Category Vanquish Loop Heart-Cut 2D-LC Systems
System pressure, maximum 700, 1000 or 1500 bar, depends on system configuration
Flow range 0-5, 0-8 or 0-10 mL/min, depends on pumps selected
Pump options (default delay volume)
Binary (35, 200 or 400 µL)
Quaternary (679 µL)
Dual (679 µL)
Isocratic
Delay volume depends on pump(s) selected
Biocompatible Yes, depends on system configuration
pH range
2–12, chloride concentration up to 1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) Yes, depends on system configuration
Solvent monitoring system compatible (optional) Yes, depends on system configuration
Customizable gradient delay volume to simplify method transfer Yes, depends on system configuration
SmartInject technology for maximum injection precision and improved column lifetime Yes
Injection volume range
Standard: 0.01-25 or 0.01-100 µL (Vanquish Flex/Vanquish Horizon)
Optional: Up to 0.01-100 or 0.01-1000 µL (Vanquish Core)
Depends on sampler selected
Sample cooling
Yes, 4-40°C 
Depends on sampler selected
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials (≤ 1.5mL) each, other rack formats available
+ capacity of 12 x 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available Yes, with Vanquish Charger
Column compartment thermostatting
Yes, 5-120°C, depends on column compartment selected
Still air and forced air
Column compartment pre-heater
Active and passive  
Depends on system configuration
Column compartment post-column cooler
Yes
Depends on column compartment selected
Detector choices
Vanquish Charged Aerosol Detector
Vanquish Diode array Detector
Vanquish Fluorescence Detector
Vanquish Multiple Wavelength Detector
Vanquish Variable Wavelength Detector
Refractive Index Detector
Mass Spectrometer
Component specifications
Standard Vanquish Loop Heart-Cut 2D-LC systems are made up of the following components. Download the associated specifications sheet for more detail on each component. 
  Vanquish Horizon Loop Heart-Cut 2D-LC System 
Vanquish Flex Loop
Heart-Cut 2D-LC System
Vanquish Core Loop Heart-Cut 2D-LC System
Pump
Download Specification Sheet
Vanquish Binary Pump H
Vanquish Binary Pump F
or 
Vanquish Quaternary Pump F
or 
Vanquish Binary Pump F
and 
Vanquish Quaternary Pump F
or 
Vanquish Dual Pump F
Vanquish Binary Pump C
or 
Vanquish Quaternary Pump C
or 
Vanquish Binary Pump C
and 
Vanquish Quaternary Pump C
or 
Vanquish Dual Pump C
Autosampler
Download Specification Sheet
Vanquish Split Sampler HT Vanquish Split Sampler FT Vanquish Split Sampler CT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger
(optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment H Vanquish Column Compartment C
Virtually zero dead volume system connections
Download Specification Sheet
Viper Fractionation Loop MP35N: 6823.0030 (250 µL), 6823.0029 (50 µL)
Viper Fractionation Loop MP35N: 6823.0030 (250 µL), 6823.0029 (50 µL)
nanoViper Fractionation Loop, PEEK shielded fused silica  
6826.2420 (20µL), 6826.2450 (50µL), 
6826.2412 (125µL)
Viper Fractionation Loop SST: 6830.2418 (200 µL) 6830.2416 (50 µL) 6830.2416 (20 µL)
Detectors
Visit our HPLC and UHPLC Detectors page
Contact us to get a quote or discuss an HPLC/UHPLC solution
1x1 image pixel for data collection